Automated and accelerated method development
Accelerate and automate your method development using Thermo Scientific Vanquish Method Development HPLC and UHPLC systems. These systems can optimize separation conditions by testing different combinations of buffers on up to six HPLC columns. Vanquish Method Development HPLC and UHPLC systems use Thermo Scientific Chromeleon Chromatography Data System (CDS), which features downloadable eWorkflows to simplify automated column and buffer scouting while enabling secure and central data storage. Achieve fully automated method development with ChromSwordAuto Chromeleon Connect and ChromSword AutoRobust Chromeleon Connect. Chromeleon CDS is also compatible with the S-Matrix® Fusion QbD® software package.
Highlights
Automated column and eluent screening option
Screen up to 13 eluents by equipping your Vanquish Method Development HPLC and UHPLC system with a solvent selection valve for extensive, automated screening. The Thermo Scientific Viper Method Scouting Kit contains all the tubing and near-zero dead volume connections to screen four columns automatically without the need for any user interaction.
Universal and sensitive detection techniques
Detect all non- or semi-volatile analytes using a near-universal Thermo Scientific Charged Aerosol Detector. Additionally, gain more confidence in identification using the Thermo Scientific ISQ EC and EM single quadrupole mass detectors for peak identification and impurity profiling.
Automated, software-assisted method development
Automate, simplify and accelerate method development. The Vanquish Method Development HPLC and UHPLC systems are controlled by Chromeleon CDS and compatible with leading method development software packages, such as ChromSwordAuto Chromeleon Connect or Fusion QbD software.
Secure data storage
The Chromeleon Data Vault ensures secure data storage and accessibility for users within the CDS infrastructure. With Vanquish Method Development HPLC and UHPLC systems, all data generated is safely backed up in Chromeleon CDS Data Vault, enabling easy data exchange throughout the organization.
Optimized for small and large molecules
When built using Vanquish Flex and Vanquish Horizon UHPLC components, these systems are biocompatible, enabling robust performance, even with high-salt gradients, while minimizing unwanted interactions between the analyte and system components. ChromSwordAuto Chromeleon Connect includes dedicated algorithms for more efficient separations of large biomolecules.
Method validation
Automate design and execution of robustness testing using ChromSword AutoRobust Chromeleon Connect and accelerate your method validation using the workflows available in the Chromeleon 7 Extension Pack ICH templates.
What are the stages of HPLC method development?
  1. Method scouting includes screening column and mobile phase conditions to choose the most promising combination for a successful HPLC separation. Traditionally, method scouting requires a significant amount of effort for manual column and mobile phase switching as well as instrument method creation. This process can be automated and streamlined using Viper fingertight fittings and solvent extension kits, and software packages such as ChromSwordAuto Chromeleon Connect and Fusion QbD.
  2. Method optimization, involving iterative testing of various separation conditions of the HPLC method, is performed to achieve the best possible resolution, speed, etc. Operators must run many samples and repeatedly make decisions based on results. This is the most time-consuming stage of method development and requires a great deal of user experience and knowledge. ChromSwordAuto Chromeleon Connect and Fusion QbD software significantly accelerates this step by rapidly analyzing separation data and automatically optimizing the HPLC method.
  3. Robustness testing is required to determine the impact of parameter variation on separation quality and is crucial for method validation. DOE and robustness testing can be automated using ChromSword AutoRobust Chromeleon Connect or Fusion QbD software.
  4. Method validation is the industry-specific process for determining whether a method is fit for purpose. Secure documentation of the method development process is necessary in a variety of industries. Chromeleon CDS ICH validation templates and documentation of all stages of the workflow can be stored in the secure Chromeleon Data Vault to ensure compliance.
Videos
System specifications
The table below provides an overview of the system specifications for the Vanquish Method Development HPLC and UHPLC systems for a standard setup. Not all specifications are listed. Your specifications may differ based on the components of your system. Find the specification sheets for the system components in the Component specifications, below.
Category
Vanquish Method Development HPLC and UHPLC Systems
System pressure, maximum
700, 1000 or 1500 bar, depends on system configuration
Flow range
0-5, 0-8 or 0-10 mL/min, depends on pumps selected
Pump options (default delay volume)
Binary (35, 200 or 400 µL)
Quaternary (679 µL)
Dual (679 µL)
Isocratic
Delay volume depends on pump(s) selected
Biocompatible
Yes, depends on system configuration
pH range
2-12, chloride concentration up to 1 mol/L
or 1-13 (chloride concentration up to 0.1 mol/L), 
depends on system configuration
System design
Modular
Touchscreen user interface display compatible (optional)
Yes, depends on system configuration
Solvent monitoring system compatible (optional)
Yes, depends on system configuration
Customizable gradient delay volume to simplify method transfer
Yes, depends on system configuration
SmartInject technology for maximum injection precision and improved column lifetime
Yes
Injection volume range
Standard: 0.01-25 or 0.01-100 µL (Vanquish Flex/Vanquish Horizon)
Optional: Up to 0.01-100 or 0.01-1000 µL (Vanquish Core)
Depends on sampler selected
Sample cooling
Yes, 4-40°C 
Depends on sampler selected
Sample capacity (standard)
Based on standard 12 mm OD vials (≤ 1.5 mL)
4 racks of 54 vials (≤ 1.5mL) each, other rack formats available
+ capacity of 12 x 22.5 mm OD vials (≤ 10 mL) in the carousel
Sampler extension available
Yes, with Vanquish Charger
Column compartment thermostatting
Yes, 5-120°C, depends on column compartment selected
Still air and forced air
Column compartment pre-heater
Active and passive  
Depends on system configuration
Column compartment post-column cooler
Yes
Depends on column compartment selected
Detector choices
Vanquish Charged Aerosol Detector
Vanquish Diode array Detector
Vanquish Fluorescence Detector
Vanquish Multiple Wavelength Detector
Vanquish Variable Wavelength Detector
Refractive Index Detector
Mass Spectrometer
Component specifications
Standard Vanquish Method Development HPLC and UHPLC systems are made up of the following components. Download the associated specifications sheet for more detail on each component.
Vanquish Horizon Method Development UHPLC System 
Vanquish Flex Method Development UHPLC System
Vanquish Core Method Development HPLC System
Pump
Download Specification Sheet
Vanquish Binary Pump H
 Vanquish Binary Pump F 
or 
Vanquish Quaternary Pump F 
or 
Vanquish Dual Pump F
Vanquish Binary Pump C 
or 
Vanquish Quaternary Pump C 
Autosampler
Download Specification Sheet
Vanquish Split Sampler HT or
Vanquish Dual Split Sampler HT
Vanquish Split Sampler FT
or
Vanquish Dual Split Sampler FT
Vanquish Split Sampler CT
or
Vanquish Split Sampler C
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger  
(optional)
Column compartment
Download Specification Sheet
Up to 3 Vanquish Column Compartment H
or
Up to 3 Vanquish Column Compartment C
Virtually zero dead volume system connections - Method scouting capabilities
Download Specification Sheet
Automated Viper Method Scouting kit for Vanquish Systems (6036.2807)
+
2 x Valve 6 position 7 port (6036.1570)
Automated Viper Method Scouting kit for Vanquish Systems (6036.2807)
+
2 x Valve 6 position 7 port (6230.1530)
Virtually zero dead volume system connections - Eluent scouting capabilities
Extension kit for Automated Method Scouting
(6036.0100)
Detectors
Visit our HPLC and UHPLC Detectors page
ChromSword is a trademark of VWR International BVBA. S-Matrix and Fusion QbD are trademarks of S-Matrix Corporation.
Contact us to get a quote or discuss an HPLC/UHPLC solution