Maximize your ICP-MS productivity with workflow-driven software and quality consumables
Discover an extensive range of products, including all the items required to support instrument uptime – nebulizers, spray chambers, torches, sample introduction systems, and more.
Qtegra ISDS Software
The Thermo Scientific Qtegra Intelligent Scientific Data Solution (ISDS) Software platform enables the analysis of trace elements by ICP-MS. The Qtegra ISDS Software streamlines and automates your workflow, enables extensive control for optimizing analytical conditions, and provides the flexibility required for the most demanding applications.
Sample and skimmer cones
A range of easily changeable Ni and Pt Thermo Scientific sample and skimmer cones is available to suit your analysis requirements. All cones are designed for durability, performance and ease of use. The cones and accessories listed below are compatible with the Thermo Scientific iCAP MX Series ICP-MS.
Spray chambers
Available in glass or PFA to suit your analysis requirements, these Thermo Scientific Cyclonic Spray Chambers efficiently filter out larger aerosol droplets for improved plasma stability. The spray chambers and accessories listed below are compatible with the iCAP MX Series ICP-MS.
Torches
All iCAP MX Series ICP-MS torches provide excellent plasma stability and are easy to install with a simple connection-free gas supply. All connectivity is contained within the torch holder to reduce complexity. The torches and accessories listed below are compatible with the iCAP MX Series ICP-MS.
Catalog # Name Price (USD) Qty
1230790
395.00
BRE0013019
436.00
BRE0024760
5,430.00
1231000 -
Injectors
Our reliable, screw-in, self-aligning injectors are available in a variety of internal diameters and materials, so you can choose the most appropriate option for your sample matrices and ICP-MS analysis needs. The injectors listed below are compatible with the iCAP MX Series ICP-MS.
Nebulizers
The nebulizers for the iCAP MX Series ICP-MS are available with different flow rates to suit your analytical needs. The nebulizers listed below are compatible with the iCAP MX Series ICP-MS.
Peristaltic pump tubing
The different pump tubing types listed below are compatible with the iCAP MX Series ICP-MS.
SEM detector for iCAP MX Series ICP-MS systems
All ICP-MS detectors have a limited lifetime based on usage. The replacement detector below is compatible with the iCAP MX Series ICP-MS.
Sample handling kits
The following sample handling kits are designed to ensure that iCAP MX Series ICP-MS systems can be configured for specific applications. The sample handling kits below are compatible with the iCAP MX Series ICP-MS.
Autosampler racks (iSC-65, ESI, Cetac)
The different autosampler racks listed below are for use with autosamplers compatible with the iCAP MX Series ICP-MS.