Protein Crosslinking
Crosslinkers contain reactive ends to specific functional groups, such as primary amines and sulfhydryls on proteins and antibodies, enabling the covalent joining of two or more biomolecules. Homobifunctional crosslinkers have the same reactive chemistry at both ends and heterobifunctional crosslinkers have different chemistries at each end. PEGylated crosslinkers are also available which provide enhanced solubility, increased stability, reduced aggregation, and reduced immunogenicity to proteins.
Use our Crosslinker Selection Tool to find the optimal protein crosslinker reagent for your protein function or interaction application, or access our helpful Bioconjugation Technical Handbook to help improve your crosslinking results.
Crosslinker Selection Tool
What is protein crosslinking?
Crosslinkers, also known as bifunctional crosslinkers, are reagents that contain two or more reactive groups which covalently attach via a spacer to functional groups on proteins or other biomolecules. Three types of crosslinkers are available: homobifunctional crosslinkers, heterobifunctional crosslinkers, and photoreactive crosslinkers. Homobifunctional crosslinking reagents have identical reactive groups, primarily amine-to-amine or sulfhydryl-to-sulfhydryl. They are typically used to form intramolecular crosslinks or to prepare polymers from monomers. Heterobifunctional crosslinking reagents have different reactive groups such as amine-to-sulfhydryl, carboxyl-to-amine, or sulfhydryl-to-carboxyl. They are useful for preparing conjugates between two different biomolecules. Heterobifunctional crosslinking reagents also include photoreactive crosslinking reagents that react with nucleophiles or form C-H insertion sites after exposure to UV light. Learn more about crosslinking reagents.
Find the optimal crosslinking reagent for your application using our Crosslinker Selection Tool or select from the most common crosslinkers below.
Most common crosslinkers based on type and reactive chemistry
Amine-to-amine crosslinkers
Reactive groupOrder productsSpacer Arm (Å)Cleavable?Water-soluble?Membrane-permeable?Reversible?
NHS esterDSG7.7N/ANoYesNo
DSS  11.4N/ANoYesNo
BS311.4N/AYesNoNo
BS(PEG)9  35.8N/AYesNoNo
DSP  12.0ThiolsNoYesNo
DTSSP  12.0ThiolsYesNoNo
EGS  16.1HydroxylamineNoYesYes
Sulfhydryl-to-sulfhydryl crosslinkers
Sulfhydryl -to-Sulfhydryl CrosslinkersOrder productSpacer Arm (Å)Cleavable?Water-soluble?Membrane permeable?
NHS-MaleimideSMCC8.3N/ANoYes
Sulfo-SMCC8.3N/AYesNo
NHS-Pyridyldithiol CrosslinkersSPDP6.8ThiolNoYes
LC-SPDP15.7ThiolNoYes
Amine-to-sulfhydryl crosslinkers
Reactive groupOrder productSpacer Arm (Å)Cleavable?Water-soluble?Membrane permeable?
NHS-Maleimide
SMCC8.3N/ANoYes
Sulfo-SMCC8.3N/AYesNo
NHS-Pyridyldithiol CrosslinkersSPDP6.8ThiolNoYes
Sulfo-LC-SPDP15.7ThiolYesNo
Carboxyl-to-amine crosslinkers
Reactive groupOrder productCleavable?Water-soluble?Membrane permeable?
Carbodiimide / NHS ester
EDCNoYesNo
NHS  NoNoYes
Sulfo-NHSNoYesNo
Photoreactive crosslinkers
Reactive groupOrder productSpacer Arm (Å)Cleavable by?Water-soluble?Membrane permeable?
NHS ester/ aryl azide
Sulfo-SANPAH18.2 LongNoYesNo
NHS ester/ diazirineSDA3.9 ShortNoNoYes
Sulfo-SDA3.9 ShortNoYesNo
LC-SDA  12.5 MidNoNoYes
Sulfo-LC-SDA12.5 MidNoYesNo
SDAD  13.5 MidThiolsNoYes
Sulfo-SDAD13.5 MidThiolsYesNo
Heterobifunctional kits
Heterobifunctional kits are offered to conjugate and purify a protein with a reactive maleimide or DBCO label for reaction with a sulfhydryl or azide, respectively.
Reactive groupOrder productSpacer Arm (Å)Labeling scaleNumber of reactionsLabeled protein reactive towards
TFP Ester/DBCODBCO Protein Labeling Kit17.90.1–0.5 mg5Azides
NHS-MaleimideMaleimide Protein Labeling Kit8.30.1–1 mg5Sulfhydryls
Controlled Protein-Protein Crosslinking Kit11.6
2.8
0.5-5 mg1Amine Sulfhydryl
* Not determined
Amine-to-amine crosslinkers
Reactive groupOrder productsSpacer Arm (Å)Cleavable?Water-soluble?Membrane-permeable?Reversible?
NHS esterDSG7.7N/ANoYesNo
DSS  11.4N/ANoYesNo
BS311.4N/AYesNoNo
BS(PEG)9  35.8N/AYesNoNo
DSP  12.0ThiolsNoYesNo
DTSSP  12.0ThiolsYesNoNo
EGS  16.1HydroxylamineNoYesYes
Sulfhydryl-to-sulfhydryl crosslinkers
Sulfhydryl -to-Sulfhydryl CrosslinkersOrder productSpacer Arm (Å)Cleavable?Water-soluble?Membrane permeable?
NHS-MaleimideSMCC8.3N/ANoYes
Sulfo-SMCC8.3N/AYesNo
NHS-Pyridyldithiol CrosslinkersSPDP6.8ThiolNoYes
LC-SPDP15.7ThiolNoYes
Amine-to-sulfhydryl crosslinkers
Reactive groupOrder productSpacer Arm (Å)Cleavable?Water-soluble?Membrane permeable?
NHS-Maleimide
SMCC8.3N/ANoYes
Sulfo-SMCC8.3N/AYesNo
NHS-Pyridyldithiol CrosslinkersSPDP6.8ThiolNoYes
Sulfo-LC-SPDP15.7ThiolYesNo
Carboxyl-to-amine crosslinkers
Reactive groupOrder productCleavable?Water-soluble?Membrane permeable?
Carbodiimide / NHS ester
EDCNoYesNo
NHS  NoNoYes
Sulfo-NHSNoYesNo
Photoreactive crosslinkers
Reactive groupOrder productSpacer Arm (Å)Cleavable by?Water-soluble?Membrane permeable?
NHS ester/ aryl azide
Sulfo-SANPAH18.2 LongNoYesNo
NHS ester/ diazirineSDA3.9 ShortNoNoYes
Sulfo-SDA3.9 ShortNoYesNo
LC-SDA  12.5 MidNoNoYes
Sulfo-LC-SDA12.5 MidNoYesNo
SDAD  13.5 MidThiolsNoYes
Sulfo-SDAD13.5 MidThiolsYesNo
Heterobifunctional kits
Heterobifunctional kits are offered to conjugate and purify a protein with a reactive maleimide or DBCO label for reaction with a sulfhydryl or azide, respectively.
Reactive groupOrder productSpacer Arm (Å)Labeling scaleNumber of reactionsLabeled protein reactive towards
TFP Ester/DBCODBCO Protein Labeling Kit17.90.1–0.5 mg5Azides
NHS-MaleimideMaleimide Protein Labeling Kit8.30.1–1 mg5Sulfhydryls
Controlled Protein-Protein Crosslinking Kit11.6
2.8
0.5-5 mg1Amine Sulfhydryl
* Not determined
No-weigh packaging crosslinkers
Protein crosslinking no-weigh packaging formats offer quick and convenient ready-to-use solutions. This format eliminates the need to weigh out small amounts of dry compounds and eliminates the waste of unused material. Simply reconstitute and the reagent is ready to use at the desired concentration.
Premium grade crosslinking reagents
Protein crosslinking premium grade reagents are high-quality reagents formulated for applications where product integrity and risk minimalization are essential. These premium grade reagents are ideal for long-term research projects that require consistent performance. Crosslinker premium grade products provide quality assurance review, lot sample retention, change control notification, along with an extra level of analytical testing and chemical characterization.
Comparison of specifications between premium and standard grade reagents
Test parameterTest methodPremium gradeStandard grade
PurityQuantitative NMR using an internal standardYesYes
VisualColor assessmentYesYes
SolubilitySample dissolves at specified concentration in deionized water to yield a clear, colorless solutionYesYes
IdentityInfrared spectroscopyYesYes
Mass identityMass spectrometryYesNA
Water contentKarl Fischer titrationYesNA
Trace metalsInductively coupled plasma mass spectrometry (ICP-MS)YesNA
Elemental analysisCombustion analysis (values reported for C, H, N, O, and S)YesNA
Residual solvent analysisHeadspace gas chromatographyUpon requestNA
In vivo protein crosslinking
In vivo crosslinking can be used to characterize protein interactions and ligand-receptor interactions irrespective of treatment conditions. Various sizes of in vivo crosslinkers are available to target surface and intracellular proteins for analysis by different methods such as immunoprecipitation (IP), Co-IP, chromatin immunoprecipitation (ChIP), electrophoresis mobility shift assay (EMSA), western blot, immunofluorescence (IF), and immunohistochemistry (IHC).
Homobifunctional, amine-reactive, NHS-ester crosslinkers
NHS-ester crosslinkers are widely used for in vivo crosslinking. NHS-ester, amine-specific reactions are fast and highly efficient since lysine residues are abundant and easily accessible on the surface of most proteins. Homobifunctional crosslinkers use a “shotgun” type approach for identifying protein complexes since they crosslink all interacting molecules whose lysine residues are within the crosslinkers spacer length. Subsequent detection simply requires a specific antibody or probe that targets one of the molecules in the complex.
Find homobifunctional, amine-reactive, NHS-ester crosslinkers for in vivo crosslinking
  • Cell surface protein crosslinking
    • BS3 (= Sulfo-DSS), 11.4 Å spacer (Ref 7)
    • DTSSP, 12.0 Å spacer, cleavable
    • Sulfo-EGS, 16.1 Å spacer, cleavable
  • Intracellular protein crosslinking
    • DSG, 7.7 Å spacer (Ref 1, 3)
    • DSP, 12.0 Å spacer, cleavable (Ref 4, 6)
    • DSS, 11.4 Å spacer (Ref 3)
    • EGS, 16.1 Å spacer, cleavable (Ref 2, 5)
Chemical structure of homobifunctional crosslinker DSG
Positive STAT3 bands with DSG and DSS, and combined Photo-Leucine and Photo-Methionine crosslinkers compared to negative control
Comparison of several in vivo crosslinking methods. HeLa cells treated with 1% Formaldehyde (HCHO) or 1mM homobifunctional NHS-ester crosslinker (Thermo Scientific DSG and DSS) in PBS for 10 minutes before quenching. A fourth set of HeLa cells were treated and crosslinked for 10 minutes with 4mM Photo-Leucine, 2mM Photo-Methionine (Photo-AA) according to the procedure (see subsequent section). Formaldehyde-treated and NHS-ester-treated cells were quenched with 100mM glycine pH 3 and 500mM Tris pH 8.0, respectively for an additional 15 minutes. One million cells from each condition were then lysed and 10µg of each sample was heated at 65°C for 10 minutes in reducing sample buffer containing 50mM DTT and analyzed by SDS-PAGE and Western blotting with Stat3 specific antibodies (Cell Signaling). Gapdh (Santa Cruz) and beta-actin (US Biologicals) were blotted as loading controls.
Heterobifunctional, photoreactive, NHS-ester/diazirine crosslinkers
NHS-ester/diazirine crosslinkers combine NHS-ester chemistry with diazirine-based photochemistry to create crosslinking. While typical heterobifunctional crosslinkers require specific amino acid groups at correct distances, SDA crosslinkers utilize a “two-step” reaction by crosslinking one protein using the NHS-ester then UV light to activate crosslinking via the diazirine moiety to any amino acid on a second protein.
Find heterobifunctional, photoreactive, NHS-ester/diazirine crosslinkers for in vivo crosslinking
  • Cell surface protein crosslinking
  • Intracellular protein crosslinking
    • SDA, 3.9 Å spacer
    • LC-SDA, 12.5 Å spacer
    • SDAD, 13.5 Å spacer, cleavable
Chemical structure of heterobifunctional crosslinker LC-SDA
Positive EEA1 crosslinking using SDA and SDAD (-DTT) compared to Sulfo-SDA, SDAD (+DTT), and control
Intracellular crosslinking of EEA1 protein complex using NHS-ester diazirine crosslinkers. HeLa cells (2 x 10^6) were labeled for 10 minutes with 1mM SDA, Sulfo-SDA or SDAD in PBS. Non-reacted NHS-esters were quenched with Tris•HCl, pH 8.0 at a final concentration of 100mM for 5 minutes and then rinsed with PBS. NHS-diazirine labeled cells and a mock-treated control (-) were UV irradiated using a Stratalinker 2400 at 365nm for 15 minutes at a distance of 4 cm in PBS. After UV treatment, cells were lysed with Thermo Scientific M-PER Mammalian Protein Extraction Reagent and analyzed for total protein concentration using the Thermo Scientific Pierce BCA Protein Assay. Reducing sample buffer was added to 10 µg of each sample except for a duplicate SDAD treated sample (-DTT) and separated by SDS-PAGE. Results were analyzed by Western blot using anti-EEA1 and GAPDH antibodies.
Photoreactive amino acid crosslinkers
Photo-Leucine and Photo-Methionine, used in metabolic labeling, are incorporated into proteins by protein synthesis (3). These non-toxic photo-amine acids are non-invasive and do not alter the cell’s metabolism. Photo-Leucine and Photo-Methionine are highly specific and ideal for studying hydrophobic interactions. Further, they can be used for either extracellular or intracellular crosslinking.
Find photoreactive amino acid crosslinkers for in vivo crosslinking
photoreactive crosslinkers added to cells then exposed to UV light show crosslinking by western blot analysis
Protocol summary for in vivo labeling and crosslinking using photoreactive amino acids as crosslinkers.
positive crosslinking using photoreactive amino acids versus formaldehyde
Photo-reactive amino acid and formaldehyde crosslinking are complementary techniques for protein interaction analysis. HeLa cells were mock treated (Lane 1), treated with 1% formaldehyde for 10 minutes (Lane 2), or treated with Photo-Methionine and Photo-Leucine followed by UV treatment (Lane 3). Cells were lysed and 10µg of each was analyzed by SDS-PAGE and Western blotting with antibodies against HSP90, STAT3, Ku70 and Ku86. Lower panels are beta-actin (upper band) and GAPDH (lower band), which were blotted as loading controls.
Bioconjugation and crosslinking technical handbook
Free download
Bioconjugation and crosslinking technical handbook
Everything you need to bioconjugate, crosslink, biotinylate, and modify proteins and peptides.
Download
Bulk or custom product request
Need larger quantities of our protein labeling, crosslinking or modification reagents? We offer bulk and custom fill sizes.
Request a quote
For Research Use Only. Not for use in diagnostic procedures.