Barnstead water purification systems maintenance schedule
Protect your investment
To protect your investment and to consistently produce the best water quality, it's important to maintain your water purification system by replacing consumables such as cartridges, filters, UV lights, etc. on a regular basis.
Please click on your water purification system below to review the monthly maintenance checklists. To purchase a replacement consumable, click on the part number in the table and add it to your cart.
Quick links to maintenance checklists by system:
Ensure your lab is properly set up prior to receiving your water purification system—review our pre-installation checklists:
Quick links to month:  3 | 6 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
50131956Barnstead™ GenPure™ Pro UV/UF, TOC Water Purification SystemSterile 0.2µm Filter09.1003
50131948Barnstead™ GenPure™ Pro  Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131950Barnstead™ GenPure™ Pro  Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131954Barnstead™ GenPure™ Pro  Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131922Barnstead™ GenPure™ Pro Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
50131956Barnstead™ GenPure™ Pro Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131948Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
50131950Barnstead™ GenPure™ Pro Ultrapure Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
UV Ultra filter50133980
50131954Barnstead™ GenPure™ Pro Ultrapure Water Purification System with ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV Ultra filter50133980
^ Top  
Quick links to month:  3 | 6 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
^ Top  
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50136151Barnstead™ GenPure™ xCAD Plus UV/UF (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
50136165Barnstead™ GenPure™ xCAD Plus (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50136146Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
50136167Barnstead™ GenPure™ xCAD Plus UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
Ultrafilter50133980
50136169Barnstead™ GenPure™ xCAD Plus UV/UF (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
50136171Barnstead™ GenPure™ xCAD Plus UV - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
UV lamp09.2002
50136172Barnstead™ GenPure™ xCAD Plus UV/UF - TOC (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
50136149Barnstead™ GenPure™ xCAD Plus (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50136152Barnstead™ GenPure™ xCAD Plus UV (bench version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV lamp09.2002
50136170Barnstead™ GenPure™ xCAD Plus UV (wall version)Sterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
UV lamp09.2002
^ Top  
Quick links to month:  3 | 6 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131243Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50131229Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and TOC monitorSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV lamp09.2002
50131211Barnstead™ GenPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50131217Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation and ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV lamp09.2002
50131235Barnstead™ GenPure™ Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.2005
UV Light09.2002
50131243Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
50131256Barnstead™ GenPure™ Water Purification System with UV-photo-oxidation, TOC monitor, ultrafiltration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
Ultrafilter50133980
UV lamp09.2002
^ Top  
Quick links to month:  3 | 6 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
^ Top  
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
DI Cartridge09.1006
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
DI Cartridge09.1006
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
Sterile vent filter for tank 3L and 6L/hr22.0091
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge
09.1006
Sterile vent filter for tank 3L and 6L/hr22.0091
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
Sterile vent filter for tank 3L and 6L/hr22.0091
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003 
^ Top  
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132366Barnstead™ MicroPure™ Ultrapure Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132367Barnstead™ MicroPure™ Ultrapure Water Purification System with integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132368Barnstead™ MicroPure™ UF Water Purification System with ultra filtration membraneSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132369Barnstead™ MicroPure™ Ultrapure Water Purification System with ultra filtration membrane and an integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132370Barnstead™ MicroPure™ UF/UX Ultrapure Water Purification System with ultra filtration membrane and UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132373Barnstead™ MicroPure™ UV Ultrapure Water Purification System with UV-photo-oxidationSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132374Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation and integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
50132372System Description: Barnstead™ MicroPure™ Ultrapure Water Purification System with UV-photo-oxidation, ultrafiltration membrane, and integrated 6L feed water tankSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.1006
UV lamp09.2002
Ultrafilter50133981
Sterile vent filter for tank 3L and 6L/hr22.0091
^ Top  
Quick links to month:  12 | 24
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
D4632-33Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 3 holder kit, Organic freeD5022
Epure, 3 holder kitD5029
D4631Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 3 holder kit, Organic freeD5022
Epure, 3 holder kitD5029
D4641Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 4 holder kit, Tap feed, Organic freeD5023
Epure, 4 holder kit, Pretreated feed, Organic freeD50228
Epure, 4 holder kit, Tap feedD5028
Epure, 4 holder kit, Pretreated feedD50227
D4642-33Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 4 holder kit, Tap feed, Organic freeD5023
Epure, 4 holder kit, Pretreated feed, Organic freeD50228
Epure, 4 holder kit, Tap feedD5028
Epure, 4 holder kit, Pretreated feedD50227
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
D4632-33Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 3 holder kit, Organic freeD5022
Epure, 3 holder kitD5029
D4631Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 3 holder kit, Organic freeD5022
Epure, 3 holder kitD5029
D4641Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 4 holder kit, Tap feed, Organic freeD5023
Epure, 4 holder kit, Pretreated feed, Organic freeD50228
Epure, 4 holder kit, Tap feedD5028
Epure, 4 holder kit, Pretreated feedD50227
D4642-33Barnstead™ E-Pure™ Water Purification System0.2 µm final filterD3750
Epure, 4 holder kit, Tap feed, Organic freeD5023
Epure, 4 holder kit, Pretreated feed, Organic freeD50228
Epure, 4 holder kit, Tap feedD5028
Epure, 4 holder kit, Pretreated feedD50227
^ Top  
Quick links to month:  3 | 4 | 6 | 8 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
^ Top 
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
10" 0.2 um filter for reservoir06.5555
CO2 vent filter06.5002
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
10" 0.2 um filter for reservoir06.5555
CO2 vent filter06.5002
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top 
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemUltrapure polisher cartridge09.2005
Sterile 0.2µm Filter09.1003
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemUltrapure polisher cartridge09.2005
Sterile 0.2µm Filter09.1003
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132395Barnstead™ LabTower™ EDI™ 15 Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV lamp09.2002
Activated Carbon Cartridge with 5µm prefilter06.5201
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
10" 0.2 um filter for reservoir06.5555
CO2 vent filter06.5002
50132396Barnstead™ LabTower™ EDI™ 30 Water Purification SystemSterile 0.2µm Filter09.1003
Ultrapure polisher cartridge09.2005
UV lamp09.2002
Activated Carbon Cartridge with 5µm prefilter06.5201
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
10" 0.2 um filter for reservoir06.5555
CO2 vent filter06.5002
^ Top 
Quick links to month:  3 | 4 | 6 | 8 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System 20Sterile 0.2µm Filter09.1003
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20 Sterile 0.2µm Filter09.1003
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132196Barnstead™ LabTower™ TII Water Purification System 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV06.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400350135142
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20 1 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 601 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System 20Sterile 0.2µm Filter09.1003
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20 Sterile 0.2µm Filter09.1003
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
^ Top 
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132196Barnstead™ LabTower™ TII Water Purification System 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV06.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400350135142
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20 1 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 601 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132196Barnstead™ LabTower™ TII Water Purification System 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV06.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400350135142
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 401 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20 1 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 201 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 601 Micron Prefilter for 09.400306.5101
System UV Lamp OLD UV09.4002
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System (UV) 20High Purity Water Cartridge02.2850-LAB
Vent filter50135142
50132196Barnstead™ LabTower™ TII Water Purification System 40High Purity Water Cartridge02.2850-LAB
Vent filter50135142
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40High Purity Water Cartridge02.2850-LAB
Vent filter50135142
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20High Purity Water Cartridge02.2850-LAB
Vent filter50135142
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20High Purity Water Cartridge02.2850-LAB
Vent filter50135142
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60High Purity Water Cartridge02.2850-LAB
Vent filter50135142
^ Top  
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
^ Top 
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132191Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
50132196Barnstead™ LabTower™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
Vent filter50135142
50132141Barnstead™ LabTower™ TII  Water Purification System (UV) 40Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
50132193Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
Vent filter50135142
50132393Barnstead™ LabTower™ TII Water Purification System (UV) 20Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
50132394Barnstead™ LabTower™ TII Water Purification System (UV) 60Sterile 0.2µm Filter09.1003
1 Micron Prefilter for 09.400306.5101
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
High Purity Water Cartridge02.2850-LAB
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
^ Top  
Quick links to month:  4 | 8 | 9 | 12 | 18 | 24
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrVent filter50135142
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrVent filter50135142
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrVent filter50135142
^ Top  
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrSterile 0.2µm Filter09.1003
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrSterile 0.2µm Filter09.1003
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132390Barnstead™ LabTower™ RO Water Purification System RO 20L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
50132391Barnstead™ LabTower™ RO Water Purification System RO 40L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
50132392Barnstead™ LabTower™ RO Water Purification System RO 60L/hrActivated Carbon Cartridge with 5µm prefilter06.5201
Replacement UV lamp09.5002
5 µm prefilter and hardness stabilizer06.5204
1 µm filter prefilter for 9.400306.5101
Vent filter50135142
^ Top  
Quick links to month:  4 | 6 | 8 | 9 | 12 | 15 | 18 | 21 | 24
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132123Barnstead™ Pacific™ TII Water Purification System 7  Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132124Barnstead™ Pacific™ TII Water Purification System 12Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132125Barnstead™ Pacific™ TII Water Purification System 20  Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132127Barnstead™ Pacific™ TII Water Purification System 40Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
^ Top   
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
^ Top   
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132123Barnstead™ Pacific™ TII Water Purification System 7  Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132124Barnstead™ Pacific™ TII Water Purification System 12Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132125Barnstead™ Pacific™ TII Water Purification System 20  Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132127Barnstead™ Pacific™ TII Water Purification System 40Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
^ Top   
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
^ Top   
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132125Barnstead™ Pacific™ TII Water Purification System 20Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
^ Top   
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
^ Top   
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
^ Top   
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
^ Top   
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132121Barnstead™ Pacific™ TII Water Purification System 3Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Replacement UV lamp09.5002
1 µm filter prefilter for 09.400306.5101
CO2 vent filter06.5002
50132129Barnstead™ Pacific™ TII Water Purification System 3 (UV)Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132123Barnstead™ Pacific™ TII Water Purification System 7  Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132131Barnstead™ Pacific™ TII Water Purification System 7 (UV)Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132124Barnstead™ Pacific™ TII Water Purification System 12Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132132Barnstead™ Pacific™ TII Water Purification System 12 (UV)Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132125Barnstead™ Pacific™ TII Water Purification System 20  Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50131982Barnstead™ Pacific™ TII Water Purification System 20 (UV)Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132127Barnstead™ Pacific™ TII Water Purification System 40Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
50132133Barnstead™ Pacific™ TII Water Purification System 40 (UV)Sterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
High Purity Water Cartridge09.4011
System UV Lamp50139226
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
CO2 vent filter06.5002
^ Top   
Quick links to month:  4 | 6 | 8 | 9 | 12 | 1518 | 21 | 24
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
1 µm filter prefilter for 9.400306.5101
^ Top   
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top   
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemActivated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
^ Top   
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top   
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
^ Top   
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top   
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top   
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
^ Top   
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50132385Barnstead™ Pacific™ RO 3 Water Purification SystemSterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Replacement UV lamp09.5002
1 µm filter prefilter for 09.400306.5101
Vent filter50135142
50132386Barnstead™ Pacific™ RO 7 Water Purification SystemSterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132387Barnstead™ Pacific™ RO 12 Water Purification SystemSterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132388Barnstead™ Pacific™ RO 20 Water Purification SystemSterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
50132389Barnstead™ Pacific™ RO 40 Water Purification SystemSterile 0.2µm Filter09.1003
Reverse Osmosis Membrane22.0046
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
Vent filter50135142
^ Top   
Quick links to month:  3 | 4 | 6 | 8 | 9 | 12 | 15 | 18 | 21 | 24
Month 3
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 4
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
^ Top   
Month 6
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 8
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification System1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
^ Top   
Month 9
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 12
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
0129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
^ Top   
Month 15
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 18
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 21
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
550129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
^ Top  
Month 24
System Catalog #System DescriptionReplacement Part DescriptionReplacement Part Catalog #
50129688Barnstead™ Smart2Pure™ 3 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
UV lamp09.1002
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129869Barnstead™ Smart2Pure™ 3 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129870Barnstead™ Smart2Pure™ 3 UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129872Barnstead™ Smart2Pure™ 3 UV Water Purification SystemSterile 0.2µm Filter09.1003
UV lamp09.1002
RO Membrane with integrated pretreatment 3L/hr09.2003
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
0129873Barnstead™ Smart2Pure™ 6 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129874Barnstead™ Smart2Pure™ 6 UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129885Barnstead™ Smart2Pure™ 6 UV Water Purification SystemSterile 0.2µm Filter09.1003
UV lamp09.1002
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129887Barnstead™ Smart2Pure™ 6 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
UV lamp09.1002
RO Membrane with integrated pretreatment 6 L /hr09.2006
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Sterile vent filter for tank 3L and 6L/hr22.0091
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
50129888Barnstead™ Smart2Pure™ 12 Standard Water Purification SystemSterile 0.2µm Filter09.1003
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129889Barnstead™ Smart2Pure™ 12 UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129890Barnstead™ Smart2Pure™ 12 UV Water Purification SystemSterile 0.2µm Filter09.1003
UV lamp09.1002
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
50129845Barnstead™ Smart2Pure™ 12 UV/UF Water Purification SystemSterile 0.2µm Filter09.1003
Ultrafilter50133981
UV lamp09.1002
RO Membrane with integrated pretreatment 12L/hr09.2012
Ultrapure Polisher Cartridge09.1020
1 Micron Prefilter for 09.400306.5101
Activated Carbon Cartridge with 5µm prefilter06.5201
Hardness Stabilizing Cartridge06.5452
5 µm prefilter and hardness stabilizer06.5204
CO2 vent filter06.5002
^ Top   
The Lab Water Source webinar series will discuss everything from the fundamentals of buying and maintaining a lab water system to ensuring the water meets your application needs.
View webinars