Gibco Advanced Granulation Technology (AGT) media
The most advanced media format available for cell culture
With today’s high application demands for increased productivity, efficiency and safety, you need an advanced media format that exceeds traditional dry format performance and accelerates time to market with proven success in commercial products. Leave the classical dry media concerns of complicated preparation, filtration difficulties, multistep processes and lot inconsistency behind and start achieving more with Gibco Advanced Granulation Technology (AGT) media format.
Contact us   Order now  Download whitepaper
What’s new: Efficient-Pro AGT Medium 
The Efficient Pro AGT dry media format enables a simple and scalable reconstitution process
The next-generation of AGT media to achieve higher titers, ease-of-use improvements, and streamline scale up.
  • Sustain cell viability for longer bioreactor runs
  • Designed to meet metabolic needs of mAb production
  • Offers significant performance with improved protein quality
  • Chemically defined without growth factors or insulin for consistency
  • Works optimally with Efficient-Pro AGT Feed 1 or Feed 2
What is AGT?
Decorative image of Advanced Granulation Technology (AGT) granule
Because not all dry media are equal
AGT is a granular dry media format produced through a technologically advanced process which allows manufacturing of complete formulations in a variety of serum-free, protein-free and chemically-defined media in a dry format. The AGT granules dissolve rapidly for faster media preparation time versus conventional dry powder media. Furthermore, AGT is efficient by nature, in that it is a complete medium that is pH and osmolality pre-adjusted, and offers all the benefits of liquid media without the cost, storage and transportation issues.
Benefits of using AGT
AGT offers high consistency and productivity across all stages of production: from development to commercial manufacturing.
Standard DPM
Disadvantages of Standard DPM
Disadvantages:
  • Procurement from different sources
  • Vast storage space
  • Monitoring and QC services
  • Complex hydration process​ (hydrate​, dissolve​, adjust pH and osmolality​, sterile filter​)
AGT Dry Media Format
Advantages of AGT Dry Media format
Advantages:
  • Single component
  • Fast dissolution
  •  Minimal storage, monitoring, QC
  • Simple hydration process (hydrate, dissolve, sterile filter)
Incorporating AGT into your processes:
  • Enables increased productivity with reduced downtime.
  • Reduces costs by eliminating the need for multiple raw materials and filters.
  • Accelerates time to market with ease of scale-up and proven success in commercial products.
  • Helps mitigate risk with multiple production facilities.
AGT media can therefore help reduce your total cycle costs, decreasing time involved in raw material planning, procurement and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.
AGT Products
Decorative image of a variety of AGT product container sizes
The AGT process allows production of complete, complex, chemically-defined, serum-free, and protein-free media in a dry format that is easy to use and easy to scale up.
The Gibco catalog cell culture media featured below are available in AGT format off-the-shelf. Your own custom formulation can be manufactured in AGT format on a make-to-order basis. Contact us for more information. 
Ordering information
AGT videos
Advanced Granulation Technology (AGT) Overview
Overview of the most advanced media format available for cell culture - AGT. A granular dry media format produced through a technologically advanced process which allows manufacturing of complete formulations in a dry format.
Video Player is loading.
Current Time 0:00
Duration 4:38
Loaded: 0.00%
Stream Type LIVE
Remaining Time 4:38
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextAdvanced Granulation Technology (AGT) Overview
        Overview of the most advanced media format available for cell culture - AGT. A granular dry media format produced through a technologically advanced process which allows manufacturing of complete formulations in a dry format.
      2. Now Playing
        Up NextHow to Increase Titers with Gibco FunctionMAX Titer Enhancer
        How to improve titers if you have a drop-off in specific productivity late in a high-density fed-batch culture. Gibco FunctionMAX TiterEnhancer can achieve up to double titer while reducing cost per gram of protein without increasing cell density.
      3. Now Playing
        Up NextAGT Dry Media Format R&D Benefits
        AGT is a granular dry media format that enables rapid rehydration and dissolution. AGT is a complete medium that is pH and osmolality pre-adjusted. It offers all the benefits of liquid media without the cost, storage and transportation issues.
      4. Now Playing
        Up NextAGT Format for Large Scale Manufacturing | Gibco
        Discover the QC-friendly media format with fewer steps and a faster workflow: AGT Dry Media Format from Thermo Fisher. Save time, resources and money. Benefits include minimized risks and consistent scale-up from research to production.
      5. Now Playing
        Up NextRisk Mitigation Through Dual-Redundant Manufacturing Facilities
        The video provides an introduction to Thermo Fisher Scientific’s new high-tech advanced granulation technology (AGT) media manufacturing facility in Scotland.
    Bioprocessing resources
    Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
    For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
    For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.