banner-diseases-bovine
Bovine/cattle diseases
Pasteurella multocida , Mannhemia haemolytica, and histophilus somni are important causes of bacterial pneumoniae. When a calf’s immune function is impaired by stress or diseases such as bovine rhinotracheitis virus (IBR), parainfluenza virus (PI-3), or bovine respiratory syncytial virus (BRSV), these bacterias can invade and cause disease.
Epidemiology
Bluetongue, or catarrhal fever, is caused by a double-stranded RNA virus of the genus Orbivirus and family Reoviridae. It is a noncontagious disease transmitted by insects to wild and domesticated ruminants, especially sheep.
Signs 
Seven or eight days after infection, sheep develop acute signs—high temperature, lethargy, and self-isolation from the herd. Shortly after the rise in temperature, the buccal mucosa becomes red and swollen, and large volumes of foamy saliva are produced. The tongue swells up and in some cases turns blue (hence the name of the disease). The crown of the unguis becomes red and painful. Affected animals can limp and ewes may abort. In most cases, growth is retarded and there is coat loss. Severely affected sheep may die eight to 10 days into the infection.
Regulatory requirements vary by country; products may not be available in your geographic area.
Top
Overview
BCV is an enveloped, single-stranded, positive-sense RNA virus belonging to the Coronaviridae family that causes intestinal and respiratory infections in ruminants worldwide. Infection can cause winter dysentery in adult animals, as well as calf enteritis and enzootic pneumonia complex in calves. Bovine coronavirus is transmitted via the oral-fecal or respiratory routes, and infected animals will typically shed the virus in their feces, particularly during parturition.
BCV may also reproduce in the upper respiratory tract, causing reoccurring respiratory or gastrointestinal infection in an animal. Infected wild ruminants have the potential to transmit the disease to domestic ruminants.
Signs
BCV signs may include diarrhea, tarry stool, indigestion, reduced appetite, weight loss, anorexia, weight loss depression, and dehydration. Additionally, BCV may present various respiratory signs such as nasal discharge, coughing, shortness of breath (dyspnea), and rapid breathing (tachypnea).
Bovine Herpesvirus 4 (BoHV-4) is a gammaherpesvirus ubiquitous of bovine. It belongs to the Rhadinovirus genus.
Some strains are implicated in ocular and respiratory affections (conjunctivitis, rhinitis, tracheitis, pneumonia). Other strains have been isolated in genital affections (orchitis, epididymitis, vaginitis, abortion, metritis, mastitis) and in disorders associated with the digestive tract (glossitis, enteritis, tumors of the rumen). Distribution is worldwide and the virus infects a range of ruminants, including bison, buffalo, sheep, and goats.
Top
PCR
Catalog # Name Size Price (EUR)
BHV450 VetMAX™ BHV Type 4 Kit Each
Epidemiology
Mycoplasmas—most of which are host-specific—cause chronic diseases that progress slowly in humans and animals. The bacterium Mycoplasma agalactiae is a common pathogen of small ruminants and is of major importance in veterinary medicine. In ovines this disease is always due to M. agalactiae, but other Mycoplasma species, M. mycoides and M. capricolum, can cause a similar disease in goats.
Signs
Mycoplasma agalactiae is the causative agent of contagious agalaxia, the main form of mycoplasmosis in European ovines and caprines that has a serious impact on the dairy industry. Antibiotics often fail to eradicate these infections, and in food animals mycoplasmosis is responsible for substantial economic losses.
PCR
Epidemiology
Bovine respiratory syncytial virus is a pneumovirus belonging to the Paramyxoviridae family. It is closely related to human RSV, which often infects the airways of children. Both are single-stranded RNA enveloped viruses. In bovines, RSV causes respiratory infections in young animals and dairy cows. The virus is mainly present in the lower airways (the lobes of the lungs), where it damages ciliated epithelial cells that normally protect the lung against microbial invasion. RSV infection often leads to secondary bacterial infection, notably with Pasteurella haemolytica and Corynebacterium pyogenes.
Signs 
The signs of the disease are hyperthermia, coughing, nasal and ocular discharge, anorexia, and dyspnea (if the disease is progressing towards fatal pneumonia). In dairy cows, milk production falls off.
Transmission 
RSV is transmitted by direct contact through nasal secretions, and its spread is favored by proximity, for example in winter stabling, and as a result of licking in the milking room.
Regulatory requirements vary by country; products may not be available in your geographic area.
Top
PCR
Overview
Bovine tuberculosis is a respiratory disease caused by the bacterium Mycobacterium bovis. It is a major infectious disease found worldwide in domestic animals, particularly cattle, as well as in certain wildlife populations.
Airborne transmission is the primary pathway for infection of M. bovis within and between species; however, animals may also become infected if they ingest large quantities of the bacterium.
Signs
Bovine tuberculosis is predominantly a respiratory disease affecting the lungs and associated lymph nodes. Infection is often subclinical, while clinical signs, when present, are not specifically distinctive of the disease. Symptoms may include physical weakness, anorexia, emaciation, enlargement of lymph nodes, and coughing, particularly in advanced cases of bovine tuberculosis.
Regulatory requirements vary by country; products may not be available in your geographic area.
PCR
Catalog # Name Size Price (EUR) Qty
MTBC VetMAX™ M. tuberculosis Complex PCR Kit Each
1.992,00
Epidemiology 
The BVD virus is a small, single-stranded RNA virus belonging to the genus Pestivirus. Two other animal viruses belong to this genus, namely the viruses that cause border disease (BD) in sheep and classical swine fever (CSF) in pigs. The virus that causes bovine viral diarrhea (BVD) was first identified in 1946. The disease is found throughout the world and its prevalence among animals exposed to the virus ranges from 30–80% in different countries and regions.
BVD impacts fertility in the herd 
The infection compromises the herd's fertility and induces the return of heat following death of the embryo when the cow is infected between the first and second months of gestation. Abortion and birth defects are possible throughout gestation.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
Overview
Brucellosis is an infectious disease caused by Brucella bacteria. Brucella species of major concern are B. abortus, primarily affecting cattle; B. melitensis, primarily affecting sheep and goats; and B. suis, primarily affecting pigs. All these Brucella species are non–host-specific and may be transmitted to other animal species or humans under appropriate conditions.
Brucellosis is commonly transmitted to susceptible animals by direct contact with infected animals or in an environment that has been contaminated with discharge from infected animals. Brucellosis is thus a herd or flock problem.
Brucellosis is a notifiable disease, and any occurrence of it has to be reported to the local health authority. Depending on the species and the infection rate, different eradication programs are effective. Where incidence rates are high, vaccination programs are necessary to lower the infection rate. Once this has been achieved, surveillance programs linked to slaughter of infected animals are introduced. These programs lead to "Brucellosis-Free" and "Officially Brucellosis-Free" status for specified regions or whole countries. In Europe, surveillance is regulated in the EU Directive 2003/99/EC on monitoring of zoonoses.
Signs
Symptoms of a Brucella infection are often decreased milk production, weight loss, abortion, infertility, and lameness. Brucella uptake occurs orally and via skin wounds or mucus membranes. Brucella bacteria are mainly excreted with aborted fetal tissue and placenta, and with semen and milk.
Occasionally, animals may recover after a period of time. More commonly, however, the symptoms disappear but the disease prevails. Such asymptomatic animals are dangerous sources of infection.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
The genus Campylobacter is composed of Gram-negative spiral-shaped bacteria. Campylobacter fetus includes the species Campylobacter fetus subsp venerealis (CFV) and C. fetus subsp. fetus (CFF). CFV is the causative agent of bovine genital campylobacteriosis. In female cattle, typical signs are frequent returns to estrus, prolonged intervals between estrus, prolonged intervals between calving and, rarely, abortions in early pregnancy.
In bulls, the contamination occurs without causing symptoms, but the infectious agent often survives for years in the lining of the foreskin.
The infection can be transmitted during intercourse with infected semen. This infection can cause significant economic losses because it causes fertility problems and infectious abortions in domestic sheep, goats, and cattle.
Bovine genital campylobacteriosis is a notifiable disease (2012 OIE List) and is considered to have socio-economic implications for public health and, in particular, international trade in animals and products of animal origin.
Top
PCR
Catalog # Name Size Price (EUR) Qty
CFP50 VetMAX™ C. fetus Kit Each -
In ruminants, chlamydiosis is a contagious disease caused by a bacterium that also infects birds and humans. It can cause abortion, premature delivery, pneumonia, conjunctivitis, and arthritis in ruminants. The infection can be transmitted to the newborn, young, or adult ruminant by the large numbers of bacteria excreted in fetal envelopes and fluids, feces, urine, and milk, although the main route of infection is via the inhalation of contaminated aerosols. Chlamydia taxonomy was revised by Everett in 1999. In the new classification, the species are divided between two main genera:
  • The genus Chlamydia, which includes C. trachomatis (humans), C. suis (pigs), and C. muridarum (mice and hamsters)
  • The genus Chlamydophila, which consists of six species, namely Cp. abortus (mammals), Cp. psittaci (birds), Cp. felis(cats), Cp. caviae (guinea pigs), Cp. pecorum (mammals), and Cp. pneumoniae (humans)
In ruminants, two species have been identified: Cp. abortus (which causes abortion) and Cp. pecorum (which causes asymptomatic gut infection, pneumonia, conjunctivitis, arthritis, abortion, and infertility). In pregnant women, C. abortus can also induce abortion with serious complications.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
Epidemiology
Q (Query) fever is an ubiquitous zoonosis that is found throughout the world (apart from New Zealand). It is caused by Coxiella burnetii, an obligate intracellular bacterium that can infect many different animal species, including ruminants, dogs, cats, birds, and arthropods as well as humans. In ruminants (which are believed to be the main reservoir for human infection), the disease is mainly associated with reproductive dysfunction. It usually remains asymptomatic and is not usually screened for unless an animal has aborted several times or shows reproductive problems. Coxiella burnetii colonizes the placenta and causes premature delivery, low birth weight, and abortion.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Signs
Signs of cryptosporidiosis can include diarrhea that persists for several days, significant weight loss, emaciation, apathy, anorexia, and dehydration.
Human health risk
Cryptosporidium can be a common nonviral cause of diarrhea in immunocompetent persons (e.g., children) and can have a severe health impact on immunocompromised persons. Infected animals can transmit the disease directly to humans, and there is also a risk of cryptosporidiosis being transmitted through surface and drinking water that has been contaminated with the feces of an infected animal.
Epidemiology
EHD is a vector-borne infectious disease caused by a virus belonging to the Orbivirus genus of the Reoviridae family (like the bluetongue virus). Between eight and 10 different serotypes have been identified.
Transmission
The virus is more-or-less exclusively transmitted by the bite of a small hematophagous dipteran of the genus Culicoides in the Ceratopogonidae family. The vector becomes infected when it feeds on an infected animal and then the virus replicates until it reaches the density necessary for transmission to another susceptible animal.
Signs
The EHD virus can infect bovines and cause signs similar to those of bluetongue (i.e., fever, erosive lesions and ulcers of the oropharyngeal mucosae, stiffness, collapse, and cutaneous edema). In gestating cows, infection can induce abortion or hydranencephaly if the infection appears between 70 and 120 days of gestation.
Regulatory requirements vary by country; products may not be available in your geographic area.
Top
PCR
Catalog # Name Size Price (EUR) Qty
EHDV50 VetMAX™ EHDV Kit Each -
Overview
FMD is a highly contagious viral disease that affects all cloven-hoofed animals, and is widespread throughout the world. FMD cannot be differentiated clinically from other vesicular diseases such as swine vesicular disease (SVD).
The virus is a member of the genus Apthovirus in the family Picornaviridae. There are seven serotypes of FMD virus: O, A, C, SAT 1, SAT 2, SAT 3, and Asia 1. Infection with any one serotype does not confer immunity against other serotypes.
Of the domesticated species, cattle, pigs, sheep, and goats are susceptible to FMD.
Signs
Clinical signs of FMD are the appearance of vesicles (small blisters) on the feet, in and around the oral cavity, and on the mammary glands of females. Vesicles can also occur inside the nostrils and at pressure points on the limbs, especially in pigs.
Transmission generally occurs through contact between infected and susceptible animals. The virus can be excreted into the air during the acute phase of infection.
Regulatory requirements vary by country; products may not be available in your geographic area.
Top
Other
Catalog # Name Size Price (EUR) Qty
6000200 IPC Evaluation Software, MS Excel-based Each -
Epidemiology
IBR is caused by a herpes virus (BHV-1), which infects the respiratory tract, causing problems on these channels (tracheitis and rhinitis), fever, abortions and infertility. It can cause death when the disease spreads rapidly within a herd. IBR is transmitted by direct contact through respiratory, eye and genital secretions.
IBR virus has an immunosuppressive effect and little infection predispose the host to a secondary bacterial infection. The infection can be latent: although primary infection may seem healed, the virus is still present in the lymph nodes and can be reactivated to spread throughout the body.
Regulatory requirements vary from country to country; it is possible that some products are not available in your area.
Top
ELISA
PCR
Catalog # Name Size Price (EUR) Qty
4415203 VetMAX™ IBR/BHV-1 Reagents Each
606,00
Overview
Leptospirosis occurs worldwide, and while the disease is typically reported in tropical climates, it can also be found in temperate climates during periods of rainfall. Leptospirosis most commonly affects pigs, cattle, and horses, and displays a variety of clinical effects ranging from mild infection to organ failure to death.
The disease is prevalent in wild mammals, but is often only noticed when the wildlife serve as an infection source for domestic animals and livestock. Leptospirosis can be transmitted transplacentally or venereally, but most often through direct contact with infected milk, urine, or placental fluids.
Signs
The signs of leptospirosis can vary greatly depending upon a herd’s immunity, age of an infected animal, and the infecting serovar. In respect to infecting serovars, there are >220 pathogenic serovars of Leptospira that can cause leptospirosis.
For other Leptospira serovars such as grippotyphosa, icterohaemorrhagiae, or pomona, cattle become incidental hosts for the disease and may display various clinical signs such as lethargy, jaundice, fever, anemia, and red urine. While adult cattle do not usually die from leptospirosis, the disease can be fatal to calves and may affect pregnant cows, causing abortion, stillbirth, and the birth of weak calves.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
ELISA
PCR
Mycoplasma bovisis a member of the Mycoplasma genus of bacteria, found.. it Causes a chronic disease causing long-term losses to dairy and beef producers, and potentially resulting in severe disease and significant welfare issues. Mycoplasma bovis causes a constellation of diseases, including mastitis in dairy cows, arthritis in cows and calves, pneumonia in calves, and various other diseases likely including late-term abortion. Not all infected cows get sick – some shed the disease without becoming ill, allowing for transmission between farms if these cows are moved. Mycoplasma bovis does not cause disease in humans. It does not cause severe disease in animals besides cattle, although it can cause rare mild infections in sheep, goats, and other related species.
Top
PCR
Catalog # Name Size Price (EUR)
MPBO50 VetMAX™ M. bovis Kit Each
Epidemiology
Neospora caninum is a protozoan parasite first observed in dogs, in which it causes myositis and encephalitis. However, in the 1990s it was observed that Neospora was a major cause of abortion in cows, usually between the fourth and seventh months of gestation. Depending on the number of infected cows in the herd, the abortion rate ranges from 5–30%; the higher rates are characterized by serial abortions occurring in less than a month.
Transmission
It is not fully understood how the parasite is transmitted, but the main route seems to be from mother to offspring, with at least 80% of the calves born to seropositive cows infected. In addition, it has been suggested that dogs may be involved in the transmission of Neospora to bovines. This infection is known on all continents and is the leading cause of bovine abortion in some countries (ahead of BVD and IBR).
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
PCR
Catalog # Name Size Price (EUR) Qty
NEOSP LSI VetMAX™ Neospora caninum Detection Kit Each
1.302,00
Epidemiology
Parainfluenza virus type 3 (PI3) is an RNA virus belonging to the Paramyxoviridae family, which is extremely common in livestock throughout the world. PI3 has been detected in both respiratory and digestive conditions, and it causes respiratory problems in young bovines. PI3 is also considered a cofactor in conditions associated with infection by certain bacteria (Mycoplasma bovis and Pasteurella haemolytica), and other viruses (including those that cause BVD and IBR).
Signs
The signs of the disease are hyperthermia, coughing, ocular and nasal discharge, anorexia, dyspnea, and diarrhea. PI3 is also immunosuppressive, and infected animals are predisposed to secondary infections.
Transmission
PI3 is usually transmitted in nasal secretions, and its transmission is favored by animal transport (direct contact, poor ventilation, and a stagnant atmosphere). To confirm PI3 infection, serological testing is strongly recommended.
Regulatory requirements vary by country; products may not be available in your geographic area.
Top
PCR
Catalog # Name Size Price (EUR) Qty
TRSVPI350 VetMAX™ BRSV PI3 Kit 50 tests -
Overview
Paratuberculosis, also known as Johne’s disease, is caused by the presence of Myobacterium avium spp. paratuberculosis (sometimes abbreviated MAP) in the small intestine of ruminants. It is a worldwide animal health problem, especially affecting beef and dairy herds.
Paratuberculosis is a chronic debilitating enteritis, and its presence in an animal herd can have serious effects on production.
Diagnosis of clinical infection is usually confirmed by the demonstration of the causal organism, M. avium ssp. paratuberculosis, in feces or in intestinal tissues postmortem. The identification of subclinical disease in animals, which can shed the organism over long periods and thus be the source of infection for other members of a herd, is more difficult. The prevalence of bovine paratuberculosis in Europe varies from country to country, ranging from 7% to 55%.
Signs
Chronic inflammation of the intestine, mesenteric lymph node lesions, diarrhea, weight loss, and edema usually appear in animals of over 2 years of age with an advanced stage of the disease.
Regulatory requirements vary by country; products may not be available in your geographic area.
Find out more
Top
PCR
Overview
Bovine rotavirus is categorized within the Reoviridae viral family, and along with coronavirus, contributes to >50% of the diarrhea problems in calves. While rotavirus is common in cattle herds, other animals such as pigs, chickens, sheep, horses, and rabbits are also susceptible to the disease.
The virus enters through the small intestine, attacking villi and making it difficult for material to be effectively absorbed into the animal’s body. This condition leads to water loss from the epithelial cells and body, causing dehydration and potentially death if there is significant dehydration and emaciation.
Transmission generally occurs when an unaffected animal has oral contact with infected feces and contaminated feed, or if they are exposed to living quarters with poor hygiene characteristics. Cows displaying signs and symptoms may shed the virus for as long as a week, while some cows can become reinfected and shed the virus throughout their life and remain asymptomatic.
Signs
The primary sign of rotavirus is a runny, often yellow diarrhea that contains mucous, blood, or both. Additionally, the animal may become lethargic and have a disinterest in drinking or feeding, which could result in mild to severe dehydration, depending on the animal and the circumstances.
PCR
Catalog # Name Size Price (EUR) Qty
RRC50 VetMAX™ Ruminant Rotavirus & Coronavirus Kit Each -
Overview
Salmonellosis is an infection caused by Salmonella bacteria. The presence of salmonellosis has been identified in countries throughout the world, but it appears to be most prevalent in areas of intensive animal husbandry, especially occurring in pigs, calves, and some types of poultry.
Many animals, especially pigs, cattle and poultry, may also be infected but show no clinical illness. Such animals may be important in relation to the spread of infection between flocks and herds and also as a source of food contamination and human infections.
Signs
Typical clinical signs can include fever and severe watery diarrhea with subsequent rapid onset of dehydration. The diarrhea is usually putrid and may contain blood and mucus. Salmonellae produce toxins that can contribute to gut damage and have systemic effects. If sufficient damage occurs to the intestinal lining, the bacteria may enter the bloodstream, resulting in septicemia, and the bacteria can spread to the brain, lungs, joints, uterus (causing abortion in pregnant cows), and other organs.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
Epidemiology
The Schmallenberg virus belongs to the Bunyaviridae family, genus Orthobunyaviridae and is closely related to Akabane, Aino, and Shamonda viruses. This virus was first identified in November 2011 in Germany. It was found in several samples coming from bovine and ovine hosts showing atypical symptoms, not characteristic of known diseases at the time.
Signs
This virus induces weak clinical symptoms affecting the global health of the animal, such as hyperthermia, loss of appetite, decreased milk production, and in some cases, diarrhea. Infection of female ruminants during gestation can also result in the birth of malformed animals (e.g., hydrocephalus).
Diagnostics
Virus detection is optimally performed using the brain of an aborted fetus, but the virus can also be detected in blood, serum, and the spleen (FLI—German National Reference Laboratory).
See all tests for ruminant abortion pathogens
Ask about our solutions
Top
PCR
Catalog # Name Size Price (EUR) Qty
SBVS50 VetMAX™ Schmallenberg Virus Kit 50 tests -
Overview
Trichomoniasis (Trich) is a venereal disease that occurs in cattle worldwide and is typically characterized by infertility and abortions in cows and heifers, which contributes to extended calving intervals.
The disease is caused by Tritrichomonas foetus (T foetus) a small, motile protozoan that lives in the reproductive tract of cows and the penis sheath of bulls. The disease organism is transferred to a cow's vagina from the bull during breeding and causes infection as it migrates to the uterus resulting in a white sticky discharge from the vulva of the infected cows.
When an infected bull is naturally bred with cows, 30%–90% become infected, which may suggest the existence of strain differences as well as a variation in breed susceptibility to the disease. Bulls of any age can remain infected for an indefinite period of time, but in younger males this scenario is less likely to occur. In contrast, cows are typically free of infection within 3 months of breeding, though they are still susceptible to reinfection. T foetus may also be transmitted when the semen from infected bulls is used during artificial insemination.
Signs
The primary sign is infertility caused by embryonic death, which contributes to repeat breeding and scenarios where cows are in heat when they should be pregnant. In addition to reduced calving rates and calf crops being extended over 3-6 months and increased number of cows may be associated with nonpregnant abnormal reproductive diagnosis such as endometritis and pyometra. Bulls will display no signs of the T foetus but can shed the organism for an indefinite period of time. Diagnosis of the disease in bulls requires the collection and testing of a preputial fluid sample taken from the sheath of the bull's penis.
Regulatory requirements vary by country; products may not be available in your geographic area.
See all tests for ruminant abortion pathogens
Top
PCR
Catalog # Name Size Price (EUR) Qty
4483869 VetMAX™-Gold Trich Detection Kit Each -