Lab safety supplies
Every lab safety program should consider Breaking the Glass Habit, and switch to shatterproof, leakproof Thermo Scientific Nalgene labware wherever plastic labware is a suitable alternative for glass. Count on Nalgene labware to be professional, high quality products, and reliably low in extractables to support and protect your people and their work. In addition to rugged bottles, beakers and other general labware, choose from a variety of specialty safety products including Right-to-Know wash bottles, beta radiation shields, safety waste containers, Versi-Dry Bench protectors and more. 
Popular lab safety products
Featured lab safety supplies products
Nalgene safety labeled wash bottles use GHS compliant symbology. Choose from the classic Right-to-Know style or the newer Right-to-Understand style with full GHS printed content.
Nalgene Versi-Dry bench protectors quickly absorb and contain spilled chemicals and cushion breakable labware. Polyethylene backing prevents chemical breakthrough and protects counter tops.
Radiation Safety
Nalgene Radiation safety products are constructed from 3/8" (9mm) thick acrylic designed to block beta-emitting radioactive isotopes.  Provides an unobstructed view of research materials.
Splash Shields
Nalgene Polycarbonate splash shields protect the user while simultaneously providing a clear field of vision. Not for use with radioactive isotopes.
Waste Disposal
Choose from a variety of Nalgene waste disposal products including autoclavable biohazardous waste containers for solid biological waste and safety waste funnel systems for liquid chemical waste.
Safety Bottle Carrier
Nalgene safety bottle carriers hold and protect chemical bottles reducing the chance of breakage while providing secondary containment if a bottle does break or leak. Ideal for toting chemical bottles between labs and from the stock room.
Biotransport Carrier
Transport potentially infectious materials using the Nalgene BioTransport Carrier. Clear, impact resistant polycarbonate construction offers excellent visibility while containing and protecting contents. Sized to fit one Nalgene Unwire Rack or two Nalgene Half-Racks. Racks not included. 
Protect against leakage, breaking and contamination with Nalgene LDPE Safety Dispensing Jugs with Closure. A safe, lightweight alternative to glass jugs, these low density polyethylene jugs have permanently attached pour spouts and polypropylene screw closures.
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific RNase AWAY is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware
Related products
Start every day with dependable labware | Thermo Scientific Nalgene labware
Video Player is loading.
Current Time 0:00
Duration 1:46
Loaded: 9.26%
Stream Type LIVE
Remaining Time 1:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Resources
    Certificate of Compliance
    Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
    Start your request 
    Support
    Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
    California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.