decorative image
Advance to clinical applications with CTS iPSC media and reagents
Advancing induced pluripotent stem cell (iPSC) therapy research to clinical applications requires researchers to carefully select materials, as the quality of starting materials has a significant impact on the properties of the final stem cell therapy product.
Thermo Fisher Scientific has developed Gibco Cell Therapy Systems (CTS) products throughout the pluripotent stem cell therapy workflow, aiming to help facilitate the transition from discovery to commercial manufacturing. The company provides high-quality ancillary materials manufactured under Good Manufacturing Practices (GMP), helping ensure commercial scale production. These materials are extensively qualified, traceable, and accompanied by regulatory documentation.
  • Scalable manufacturing under GMP conditions—CTS iPSC media and reagents are manufactured in conformity with GMP for medical devices, 21 CFR Part 820, and follow USP<1043> and Ph Eur 5.2.12, and the manufacturing sites are FDA-registered and ISO 13485- and/or ISO 9001-certified. The investment in rigorous product manufacturing helps ensure the consistency, reliability, and high quality of iPSC therapy reagents in combination with the ability to scale up and accommodate changing needs.
  • Quality control testing and regulatory documentation—CTS iPSC media and reagents help minimize the risk of contamination and variability in research with extensive safety testing, including sterility, endotoxin, and mycoplasma. Full traceability for raw materials is also available, including Drug Master Files, Regulatory Support Files, and Certificates of Origin.
  • Proven use from research to clinic—An extensive selection of serum-free, xeno-free, or animal origin-free formulations with cell therapy specific intended use statements and complementary research use reagents. Our CTS products have been used in over 200 clinical trials as well as in commercially approved cell therapies including FDA-approved CAR-T therapies and the first FDA-approved therapeutic cancer vaccine.
Explore CTS iPSC media and reagents
GMP iPSC Reprogramming kit
CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit
The first off-the-shelf reprogramming system manufactured in accordance with GMP requirements, offering high-efficiency Sendai delivery of reprogramming factors.
  • Xeno-free workflow—generate iPSC lines from both fibroblasts and blood for clinical research
  • Regulatory support—Traceability documentation, such as Certificate of Analysis and Certificate of Origin, available to support regulatory filing
  • Designed with transition in mind—complimentary research-use CytoTune iPS 2.0 Sendai Reprogramming Kit helps facilitate translation to the clinic
See data
Catalog # Name Size Price (USD) Qty
A34546 CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit Each
18,900.00
A16517 CytoTune™-iPS 2.0 Sendai Reprogramming Kit, 1 Pack Each
3,933.65
Online Exclusive
4,225.00
Save 291.35 (7%)
CTS HiFi Cas9 Protein
Regulatory bodies like the FDA recognize the safety concerns of off-target cleavage events when using CRISPR-Cas9, particularly for therapeutics applications. To address these safety risks, there is a developed CTS HiFi Cas9 Protein, a GMP-manufactured high-fidelity Cas9 protein designed to significantly reduce off-target events while maintaining high editing efficiency.
  • Significantly reduce off-target effects—>90% reduction in a broad range of cell types including primary immune cells
  • Equivalent editing efficiency—Achieve the same high editing efficiency as CTS TrueCut Cas9 Protein, our wild-type protein designed for exceptional on-target editing efficiency
  • Improved knock-in efficiency—Also offered in CTS HiFi Cas9 and CTS Xenon Genome Editing Buffer Kit to offer 2X improved knock-in efficiency
See data
Order CTS HiFi Cas9 Protein
CTS TrueCut Cas9 Protein
Order CTS TrueCut Cas9 Protein
Catalog # Name Size Price (USD) Qty
A45220 CTS™ TrueCut™ Cas9 Protein, 2.5 mg Each
12,421.50
Special offer
Online exclusive
Ends: 21-Jun-2025
13,650.00
Save 1,228.50 (9%)
A45221 CTS™ TrueCut™ Cas9 Protein, 5 mg Each
26,160.00
A36496 TrueCut™ Cas9 Protein v2, 10 μg Each
90.65
Online Exclusive
92.75
Save 2.10 (2%)
A36497 TrueCut™ Cas9 Protein v2, 25 μg Each
112.65
Online Exclusive
116.00
Save 3.35 (3%)
A36498 TrueCut™ Cas9 Protein v2, 100 μg Each
240.65
Online Exclusive
248.00
Save 7.35 (3%)
A36499 TrueCut™ Cas9 Protein v2, 500 μg Each
984.65
Online Exclusive
1,022.00
Save 37.35 (4%)
iPSC media for cell therapy
CTS StemScale PSC Suspension Medium
Gibco CTS StemScale PSC Suspension Medium is designed to support the expansion of pluripotent stem cells (PSCs) at scale in suspension culture. Its scalable, easy-to-use workflow and xeno-free formulation make it exceptional for translational and clinical applications.
  • Maximal expansion capability—achieve up to 30% higher expansion than other PSC suspension media
  • Scalable expansion for clinical applications—grow PSCs across multiple culture vessel formats and sizes, from small to large scale
  • No cell strainer required—remove scale limitation, reduce contamination risk and hands-on time, and improve cell viability and yield
See data
Order CTS StemScale PSC Suspension Medium
Catalog # Name Size Price (USD) Qty
A5869601 CTS™ StemScale™ PSC Suspension Medium Each
1,545.18
Special offer
Online exclusive
Ends: 21-Jun-2025
1,698.00
Save 152.82 (9%)
A4965001 StemScale™ PSC Suspension Medium Each
992.00
CTS Essential 8 Medium
Based on the widely cited Essential 8 Medium, Gibco CTS Essential 8 Medium was the first globally available human pluripotent stem cell (hPSC) culture medium with components not directly derived from animals.
  • Supports long-term iPSC culture—Maintains iPSC marker expression and a normal karyotype over multiple passages
  • Maintains differentiation potential—PSCs cultured in CTS Essential 8 Medium are able to generate cells of all three germ layers
  • Helps reduce risk—Based on the same fully defined and highly consistent eight-component formulation as research-use Essential 8 Medium but without components directly of animal or human origin
See data
Order CTS Essential 8 Medium
Catalog # Name Size Price (USD) Qty
A2656101 CTS™ Essential 8™ Medium Each
471.00
A1517001 Essential 8™ Medium Each
339.00
CTS KnockOut SR XenoFree Medium
Order CTS KnockOut SR XenoFree Medium
iPSC reagents for cell therapy
CTS Vitronectin (VTN-N) Recombinant Human Protein
Order CTS VTN-N Recombinant Human Protein
CTS Versene Solution
Order CTS Versene Solution
Catalog # Name Size Price (USD) Qty
A4239101 CTS™ Versene™ Solution Each
43.68
15040066 Versene Solution Each
17.31
CTS KnockOut DMEM and CTS KnockOut DMEM/F12
Order CTS KnockOut DMEM and CTS KnockOut DMEM/F12
CTS RevitaCell Supplement
Gibco CTS RevitaCell Supplement (100X) is an animal-origin-free, chemically defined supplement used with PSCs for post-thaw recovery or in combination with CTS Essential 8 Medium for single cell passaging.
  • Improved cell survival—Provides high cell viability post-single cell passaging and post-thaw
  • Contains higher specificity inhibitor—Includes a higher specificity ROCK inhibitor, coupled with antioxidants and free radical scavengers to minimize impact of stress on cells
  • Offers consistency—Formulated using animal-origin free and chemically defined materials
Order CTS RevitaCell Supplement
Catalog # Name Size Price (USD) Qty
A4238401 CTS™ RevitaCell™ Supplement (100X) Each
291.00
A2644501 RevitaCell™ Supplement (100X) Each
247.00
CTS PSC Cryopreservation Kit
Order CTS PSC Cryopreservation Kit
Catalog # Name Size Price (USD) Qty
A4239301 CTS™ PSC Cryopreservation Kit Each
497.00
A2644601 PSC Cryopreservation Kit Each
425.00
CTS Essential 6 Medium
Order CTS Essential 6 Medium
Catalog # Name Size Price (USD) Qty
A4238501 CTS™ Essential 6 Medium Each
343.00
A1516401 Essential 6™ Medium Each
286.00
CTS B-27 Supplement XenoFree
Product photo
Gibco B-27 Supplement has been the trusted standard for a variety of neuronal culture applications with citations in more than 11,000 publications. The CTS™ product offers proven serum-free supplementation for the growth, expansion, differentiation, and maintenance of PSC-derived cells.
  • Applicable to numerous cell types - neural stem cells, neurons, and cardiomyocytes
  • Three varieties available - full version, minus vitamin A, and minus insulin
  • Equivalent performance - same performance as the trusted research‑use Gibco B‑27 Supplement XenoFree
Order CTS B-27 Supplement XenoFree
Catalog # Name Size Price (USD) Qty
A5047501 CTS™ B-27™ Supplement XenoFree (50X) Each
296.65
Online Exclusive
336.00
Save 39.35 (12%)
A5047601 CTS™ B-27™ Supplement XenoFree minus Vitamin A (50X) Each
305.27
Special offer
Online exclusive
Ends: 21-Jun-2025
343.00
Save 37.73 (11%)
A5047701 CTS™ B-27™ Supplement XenoFree minus Insulin (50X) Each
315.61
Special offer
Online exclusive
Ends: 21-Jun-2025
348.00
Save 32.39 (9%)
Gibco PeproTech growth factors and cytokines for iPSC therapy
GMP iPSC Reprogramming kit
CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit
The first off-the-shelf reprogramming system manufactured in accordance with GMP requirements, offering high-efficiency Sendai delivery of reprogramming factors.
  • Xeno-free workflow—generate iPSC lines from both fibroblasts and blood for clinical research
  • Regulatory support—Traceability documentation, such as Certificate of Analysis and Certificate of Origin, available to support regulatory filing
  • Designed with transition in mind—complimentary research-use CytoTune iPS 2.0 Sendai Reprogramming Kit helps facilitate translation to the clinic
See data
Catalog # Name Size Price (USD) Qty
A34546 CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit Each
18,900.00
A16517 CytoTune™-iPS 2.0 Sendai Reprogramming Kit, 1 Pack Each
3,933.65
Online Exclusive
4,225.00
Save 291.35 (7%)
CTS HiFi Cas9 Protein
Regulatory bodies like the FDA recognize the safety concerns of off-target cleavage events when using CRISPR-Cas9, particularly for therapeutics applications. To address these safety risks, there is a developed CTS HiFi Cas9 Protein, a GMP-manufactured high-fidelity Cas9 protein designed to significantly reduce off-target events while maintaining high editing efficiency.
  • Significantly reduce off-target effects—>90% reduction in a broad range of cell types including primary immune cells
  • Equivalent editing efficiency—Achieve the same high editing efficiency as CTS TrueCut Cas9 Protein, our wild-type protein designed for exceptional on-target editing efficiency
  • Improved knock-in efficiency—Also offered in CTS HiFi Cas9 and CTS Xenon Genome Editing Buffer Kit to offer 2X improved knock-in efficiency
See data
Order CTS HiFi Cas9 Protein
CTS TrueCut Cas9 Protein
Order CTS TrueCut Cas9 Protein
Catalog # Name Size Price (USD) Qty
A45220 CTS™ TrueCut™ Cas9 Protein, 2.5 mg Each
12,421.50
Special offer
Online exclusive
Ends: 21-Jun-2025
13,650.00
Save 1,228.50 (9%)
A45221 CTS™ TrueCut™ Cas9 Protein, 5 mg Each
26,160.00
A36496 TrueCut™ Cas9 Protein v2, 10 μg Each
90.65
Online Exclusive
92.75
Save 2.10 (2%)
A36497 TrueCut™ Cas9 Protein v2, 25 μg Each
112.65
Online Exclusive
116.00
Save 3.35 (3%)
A36498 TrueCut™ Cas9 Protein v2, 100 μg Each
240.65
Online Exclusive
248.00
Save 7.35 (3%)
A36499 TrueCut™ Cas9 Protein v2, 500 μg Each
984.65
Online Exclusive
1,022.00
Save 37.35 (4%)
iPSC media for cell therapy
CTS StemScale PSC Suspension Medium
Gibco CTS StemScale PSC Suspension Medium is designed to support the expansion of pluripotent stem cells (PSCs) at scale in suspension culture. Its scalable, easy-to-use workflow and xeno-free formulation make it exceptional for translational and clinical applications.
  • Maximal expansion capability—achieve up to 30% higher expansion than other PSC suspension media
  • Scalable expansion for clinical applications—grow PSCs across multiple culture vessel formats and sizes, from small to large scale
  • No cell strainer required—remove scale limitation, reduce contamination risk and hands-on time, and improve cell viability and yield
See data
Order CTS StemScale PSC Suspension Medium
Catalog # Name Size Price (USD) Qty
A5869601 CTS™ StemScale™ PSC Suspension Medium Each
1,545.18
Special offer
Online exclusive
Ends: 21-Jun-2025
1,698.00
Save 152.82 (9%)
A4965001 StemScale™ PSC Suspension Medium Each
992.00
CTS Essential 8 Medium
Based on the widely cited Essential 8 Medium, Gibco CTS Essential 8 Medium was the first globally available human pluripotent stem cell (hPSC) culture medium with components not directly derived from animals.
  • Supports long-term iPSC culture—Maintains iPSC marker expression and a normal karyotype over multiple passages
  • Maintains differentiation potential—PSCs cultured in CTS Essential 8 Medium are able to generate cells of all three germ layers
  • Helps reduce risk—Based on the same fully defined and highly consistent eight-component formulation as research-use Essential 8 Medium but without components directly of animal or human origin
See data
Order CTS Essential 8 Medium
Catalog # Name Size Price (USD) Qty
A2656101 CTS™ Essential 8™ Medium Each
471.00
A1517001 Essential 8™ Medium Each
339.00
CTS KnockOut SR XenoFree Medium
Order CTS KnockOut SR XenoFree Medium
iPSC reagents for cell therapy
CTS Vitronectin (VTN-N) Recombinant Human Protein
Order CTS VTN-N Recombinant Human Protein
CTS Versene Solution
Order CTS Versene Solution
Catalog # Name Size Price (USD) Qty
A4239101 CTS™ Versene™ Solution Each
43.68
15040066 Versene Solution Each
17.31
CTS KnockOut DMEM and CTS KnockOut DMEM/F12
Order CTS KnockOut DMEM and CTS KnockOut DMEM/F12
CTS RevitaCell Supplement
Gibco CTS RevitaCell Supplement (100X) is an animal-origin-free, chemically defined supplement used with PSCs for post-thaw recovery or in combination with CTS Essential 8 Medium for single cell passaging.
  • Improved cell survival—Provides high cell viability post-single cell passaging and post-thaw
  • Contains higher specificity inhibitor—Includes a higher specificity ROCK inhibitor, coupled with antioxidants and free radical scavengers to minimize impact of stress on cells
  • Offers consistency—Formulated using animal-origin free and chemically defined materials
Order CTS RevitaCell Supplement
Catalog # Name Size Price (USD) Qty
A4238401 CTS™ RevitaCell™ Supplement (100X) Each
291.00
A2644501 RevitaCell™ Supplement (100X) Each
247.00
CTS PSC Cryopreservation Kit
Order CTS PSC Cryopreservation Kit
Catalog # Name Size Price (USD) Qty
A4239301 CTS™ PSC Cryopreservation Kit Each
497.00
A2644601 PSC Cryopreservation Kit Each
425.00
CTS Essential 6 Medium
Order CTS Essential 6 Medium
Catalog # Name Size Price (USD) Qty
A4238501 CTS™ Essential 6 Medium Each
343.00
A1516401 Essential 6™ Medium Each
286.00
CTS B-27 Supplement XenoFree
Product photo
Gibco B-27 Supplement has been the trusted standard for a variety of neuronal culture applications with citations in more than 11,000 publications. The CTS™ product offers proven serum-free supplementation for the growth, expansion, differentiation, and maintenance of PSC-derived cells.
  • Applicable to numerous cell types - neural stem cells, neurons, and cardiomyocytes
  • Three varieties available - full version, minus vitamin A, and minus insulin
  • Equivalent performance - same performance as the trusted research‑use Gibco B‑27 Supplement XenoFree
Order CTS B-27 Supplement XenoFree
Catalog # Name Size Price (USD) Qty
A5047501 CTS™ B-27™ Supplement XenoFree (50X) Each
296.65
Online Exclusive
336.00
Save 39.35 (12%)
A5047601 CTS™ B-27™ Supplement XenoFree minus Vitamin A (50X) Each
305.27
Special offer
Online exclusive
Ends: 21-Jun-2025
343.00
Save 37.73 (11%)
A5047701 CTS™ B-27™ Supplement XenoFree minus Insulin (50X) Each
315.61
Special offer
Online exclusive
Ends: 21-Jun-2025
348.00
Save 32.39 (9%)
Gibco PeproTech growth factors and cytokines for iPSC therapy
Custom iPSC media
We offer full customization options to help meet your unique specifications for any project. Flexibility is yours in creating your own Gibco custom cell culture medium.
  • Add or remove components
  • Change a concentration
  • Switch packaging
  • Select from cGMP or non-GMP
  • Choose QC tests
Learn more
product photo
Gibco CTS Rotea Counterflow Centrifugation System
  • Process flexibility
  • High cell recovery and viability
  • Low output volumes
  • Research through commercial manufacturing
Learn more
product photo
Gibco CTS Xenon Electroporation System
  • High speed and large volume
  • Scalable, proven performance and viability
  • Process flexibility
  • Efficient nonviral transfection
  • Closed-system processing
Learn more
Featured resources for iPSC therapy
Cell therapy resources
Support
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.