fullerene
Carbon, widely distributed in nature, is the element found in all living organisms. It is among the five most abundant elements in the universe. In nature, free carbon occurs in three allotropic forms: amorphous, graphite, and diamond, with graphite being one of the softest known solids, and diamond being one of the hardest. Properties of products made from elemental carbon can be controlled by the manufacturing process and the type of carbon selected as raw material.
Shop now
The above-mentioned allotropes of carbon have been known since ancient times. More recently, two other naturally occurring forms of pure carbon have been discovered, namely fullerene and graphene for which the discoverers were awarded the Nobel Prize. Spherical fullerenes awere originally known as buckminsterfullerenes after the American architect and philosopher R. Buckminster Fuller who popularized  geodesic domes. They are also called buckyballs because they resemble the balls used in football (soccer). Fullerene itself, which consists of a hollow cluster of 60 carbon atoms, is the most symmetrical large molecule known. The atoms are connected by single and double bonds, forming a closed or partially closed mesh, with fused rings of five to seven atoms. Fullerenes with more than 60 carbons are known today. The fullerenes are also found to be soluble in common solvents such as benzene, toluene, or chloroform.
Thermo Fisher Scientific's extensive portfolio of elemental carbon products is suitable for a wide range of laboratory applications, and is also available in bulk and custom quantities and formats.
Popular elemental carbon products
Catalog # Name Price (USD) Qty
038984.03 Diamond powder, natural, 40-60 micron
90.65
Special offer
Online exclusive
Ends: 30-Jun-2025
106.00
Save 15.35 (14%)
038984.06 Diamond powder, natural, 40-60 micron
328.65
Online Exclusive
365.00
Save 36.35 (10%)
013401.03 Diamond powder, synthetic, <1 micron, 99.9% (metals basis) -
013401.06 Diamond powder, synthetic, <1 micron, 99.9% (metals basis) -
013402.03 Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis) -
013402.06 Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis) -
044508.01 Fullerene, buckytube/nanotube, single walled
59.65
Special offer
Online exclusive
Ends: 30-Jun-2025
69.70
Save 10.05 (14%)
044508.03 Fullerene, buckytube/nanotube, single walled
159.65
Special offer
Online exclusive
Ends: 30-Jun-2025
188.00
Save 28.35 (15%)
044501.02 Fullerene, buckytube/nanotube, single walled, >60% SWNT
434.65
Online Exclusive
483.00
Save 48.35 (10%)
044790.01 Fullerene, carbon nanotube, multi-walled, ∣<8 nm OD, 2-5 nm ID, 0.5-2 micron long
44.30
044790.03 Fullerene, carbon nanotube, multi-walled, ∣<8 nm OD, 2-5 nm ID, 0.5-2 micron long
90.65
Online Exclusive
101.00
Save 10.35 (10%)
044790.06 Fullerene, carbon nanotube, multi-walled, ∣<8 nm OD, 2-5 nm ID, 0.5-2 micron long
348.65
Online Exclusive
387.00
Save 38.35 (10%)
044192.03 Fullerene, nanotube, multi-walled, 20-50 nm OD, 5-20 micron long
215.00
044192.06 Fullerene, nanotube, multi-walled, 20-50 nm OD, 5-20 micron long
895.00
043839.MF Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long
38.80
043839.MD Fullerene, nanotube, multi-walled, 20 nm OD, 5-20 micron long
96.65
Special offer
Online exclusive
Ends: 30-Jun-2025
113.00
Save 16.35 (14%)
042886.04 Fullerene, nanotube, multi-walled, as-produced cathode deposits, core and shell
81.70
042886.09 Fullerene, nanotube, multi-walled, as-produced cathode deposits, core and shell
342.00
039720.MA Fullerene powder, 97% C{70} -
039720.MF Fullerene powder, 97% C{70} -
039720.MD Fullerene powder, 97% C{70} -
042601.MD Fullerene powder, 98+% C{70} -
042601.03 Fullerene powder, 98+% C{70} -
039722.MC Fullerene powder, 99.5% C{60} -
039722.03 Fullerene powder, 99.5% C{60} -
042007.MD Fullerene powder, 99.9+% C{60} -
042007.03 Fullerene powder, 99.9+% C{60} -
042600.MD Fullerene powder, 99+% C{70} -
042600.03 Fullerene powder, 99+% C{70} -
041182.MB Fullerene powder, hydroxylated, C{60}(OH)∣n,
161.65
Special offer
Online exclusive
Ends: 26-Sep-2025
190.00
Save 28.35 (15%)
041182.MC Fullerene powder, hydroxylated, C{60}(OH)∣n,
548.65
Online Exclusive
609.00
Save 60.35 (10%)
041181.MD Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2% -
041181.03 Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2% -
041181.06 Fullerene powder, mixed refined, typically 70% C{60}, 28% C{70}, higher 2% -
040970.MD Fullerene powder, mixed, typically 98% C{60}, 2% C{70} -
040970.03 Fullerene powder, mixed, typically 98% C{60}, 2% C{70} -
042008.MF Fullerene powder, sublimed, 99.9+% C{60} -
042008.MD Fullerene powder, sublimed, 99.9+% C{60} -
042008.03 Fullerene powder, sublimed, 99.9+% C{60} -
Find carbon in our interactive periodic table
Click on image to learn the physical properties of carbon or any element, as well as to select products by format or find compounds and alloys.
Search now