eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Certificates
SDS
Invitrogen™
eBioscience™ Protein Transport Inhibitor Cocktail (500X)
The Protein Transport Inhibitor Cocktail (500X) is a cocktail of Brefeldin A and Monensin. It is suitable for use inRead more
Have Questions?
Change view
Catalog Number
Quantity
00-4980-93
100 μL
00-4980-03
4 x 100 μL
2 Options
Catalog number 00-4980-93
Price (EUR)
39,93
Each
-
Add to cart
Quantity:
100 μL
Recurring order eligible. Learn more »
Price (EUR)
39,93
Each
Add to cart
Ask our AI about this Product
The Protein Transport Inhibitor Cocktail (500X) is a cocktail of Brefeldin A and Monensin. It is suitable for use in assays in which prevention of protein transport to the extracellular space is desired and has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X) (cat. 00-4975). Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus. This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 μL
Product LineeBioscience™
TypeProtein Transport Inhibitor Cocktail
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Figures
Data for Protein Transport Inhibitor Cocktail (500X)