DynaMag™-15 Magnet
DynaMag™-15 Magnet
Invitrogen™

DynaMag™-15 Magnet

The DynaMag™-15 magnet is optimized for efficient cell isolation using all types of Dynabeads™ (range 1–4.5 μm in diameter) inRead more
Have Questions?
Catalog NumberQuantity
12301D1 Each
Catalog number 12301D
Price (USD)
1,354.00
Each
-
Add to cart
Quantity:
1 Each
Recurring order eligible. Learn more »
Price (USD)
1,354.00
Each
Add to cart
Ask our AI about this Product
The DynaMag™-15 magnet is optimized for efficient cell isolation using all types of Dynabeads™ (range 1–4.5 μm in diameter) in standard 15 mL lab tubes. The DynaMag™-15 magnet holds 4 tubes and combines a strong magnetic attraction for efficient separations with a flexible and smart ergonomic design that ensures a comfortable working position. A tilting mechanism on the magnet enables flexibility while pipetting.

• Optimal working volume: 15 mL
• Holds 4 × 15 mL tubes
• Good control and visibility of your sample

For handling of smaller blood collection tubes, FACS tubes, or similar, see our DynaMag™-5 magnet.

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find other magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)DynaMag™
Quantity1 Each
Sample Volume4-15 mL
Shipping ConditionRoom Temperature
Product LineDynaMag™
TypeCell Separation Magnet
Unit SizeEach
Contents & Storage
Contains one magnet. Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

1 Tesla is equivalent to 10,000 Gauss. .

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.

Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.

This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.

Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection