1x1 image pixel for data collection
DynaMag™-96 Side Magnet
DynaMag™-96 Side Magnet
Invitrogen™

DynaMag™-96 Side Magnet

The DynaMag™-96 Side Magnet is optimized to pull all types of Dynabeads™ (range 1–4.5 μm in diameter) to the sidesRead more
Have Questions?
Catalog NumberQuantity
12331D1 Each
Catalog number 12331D
Price (USD)
1,180.65
Online Exclusive
1,236.00
Save 55.35 (4%)
Each
In stock
Add to cart
Quantity:
1 Each
Price (USD)
1,180.65
Online Exclusive
1,236.00
Save 55.35 (4%)
Each
Add to cart
The DynaMag™-96 Side Magnet is optimized to pull all types of Dynabeads™ (range 1–4.5 μm in diameter) to the sides of PCR tubes or to the sides of wells in 96-well plates. Designed for both automation platforms (e.g., Tecan) and manual workflows, this magnet is not compatible with PCR tubes but is compatible with PCR-strips and 96-well plates (200 μL), non-skirted and half-skirted.

Efficient mixing without sample handling
This magnet has an extra column (13 total). This enables sample mixing simply by shifting the 96-well plate back and forth from the right-most position to the left-most position. This shifting draws the beads back and forth through the sample and allows mixing to be achieved without handling, promoting faster processing and less sample handing-induced variation.

Handling volume
When used in combination with the DynaMag™-96 Bottom magnet on an automation platform, the two magnets allow for significant volume changes through a workflow.
• High wash volumes: DynaMag™-96 Side Magnet
• Low elution volumes: DynaMag™-96 Bottom magnet

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find other magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMagnet
For Use With (Equipment)DynaMag™
Quantity1 Each
Shipping ConditionRoom Temperature
Product LineDynaMag™
Unit SizeEach
Contents & Storage
Contains one magnet. Store at room temperature.

Frequently asked questions (FAQs)

What are the Gauss levels for the DynaMag-2 and DynaMag-Spin Magnets?

The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What is the standard conversion of Gauss to Tesla units?

1 Tesla is equivalent to 10,000 Gauss. .

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

What is the difference between the DynaMag-2 Magnet (Cat. No. 12321D) and DynaMag-Spin Magnet (Cat. No. 12320D)?

The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link