VersiPlate PCR Strip Tube Plate, 96-well, low profile
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
VersiPlate PCR Strip Tube Plate, 96-well, low profile
Thermo Scientific™

VersiPlate PCR Strip Tube Plate, 96-well, low profile

The Thermo Scientific VersiPlate, 96-well Low Profile PCR Strip Tube Plate, is unique, with each strip of eight tubes beingRead more
Have Questions?
Catalog NumberQuantity
AB180025 Plates
Catalog number AB1800
Price (USD)
164.65
Online Exclusive
188.00
Save 23.35 (12%)
Each
Add to cart
Quantity:
25 Plates
Recurring order eligible. Learn more »
Price (USD)
164.65
Online Exclusive
188.00
Save 23.35 (12%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific VersiPlate, 96-well Low Profile PCR Strip Tube Plate, is unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Features

• Easier sample handling as multiple strips of samples can be handled simultaneously
• Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
• Can be used in instruments with or without a VersiFrame.
• The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
• The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
• Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
• Maximum fill volume of 0.2 mL
• Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
VersiPlate PCR Strip Tube Plate, 96-well, low profile, white
VersiPlate Frame, 96-well, skirted, white
VersiCap Mat, 96-well, domed cap strips
VersiCap Mat, 96-well, flat cap strips
VersiCap Mat, 96-well, ultra clear cap strips
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 Plates
Reaction SpeedLow profile
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorClear
MaterialPolycarbonate
No. of Wells96
Product LineVersiPlate
Volume (Metric) Well0.2 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection