If the patient’s history indicates IgE-mediated allergy, specific IgE blood tests can assist in the diagnosis.7,8 Specific IgE blood tests can be performed at any age and use validated assays to quantify allergen-specific IgE levels in the serum.7 Firstly, a test for a specific IgE response to the whole allergen (egg) should be performed.8
Interpretation of whole-allergen specific IgE blood test results
- Results should always be interpreted in the context of the patient’s clinical history.6
- The presence of a specific IgE response to the whole allergen indicates a sensitisation to egg, and an associated IgE-mediated immunological process.8
- The higher the antibody titre, the greater the probability of having egg allergy.9
Whole-allergen tests predict the likelihood of egg allergy but are not predictive of reactivity to cooked egg. After testing for a specific IgE response to whole allergen, egg allergen component tests can help evaluate reactivity to cooked egg and provide further information on the likelihood of allergy persistence.6,8
Assessment of specific IgE responses to the following components (proteins) could help pinpoint the diagnosis and guide the management of patients with egg allergy:8
- Ovomucoid (f233 / Gal d 1)
- Ovalbumin (f232 / Gal d 2)
- Conalbumin (f323 / nGal d 3)
- Lysozyme (k208 / Gal d 4)
As ovomucoid is more resistant than other proteins to heat denaturation, patients with high levels of ovomucoid-specific IgE are likely to react to cooked egg. Such patients should therefore avoid consumption of all forms of egg. Patients who test negative for ovomucoid-specific IgE may be able to tolerate cooked egg.8
70 percent of children with egg allergy do not react to egg in baked products.10
Consider using allergen component blood tests to identify these patients.8