Name | Order | |
![]() | Platinum SuperFi II DNA PolymeaseFidelity: >300x Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Long-range PCR, Benchtop stability, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤20 kb DNA synthesis speed: 15–30 sec/kb Amplicon overhang: Blunt Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Platinum SuperFi II PCR Master MixFidelity: >300x Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Long-range PCR, Benchtop stability, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤20 kb DNA synthesis speed: 15–30 sec/kb Amplicon overhang: Blunt Product format: 2X master mix (colorless) Source: Recombinant | Order |
![]() | Platinum SuperFi II Green PCR Master MixFidelity: >300x Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Long-range PCR, Benchtop stability, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤20 kb DNA synthesis speed: 15–30 sec/kb Amplicon overhang: Blunt Product format: 2X master mix (green) Source: Recombinant | Order |
![]() | Platinum II Taq Hot-Start DNA PolymeraseFidelity: 1x (standard) Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 15 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Platinum II Hot-Start PCR Master MixFidelity: 1x (standard) Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 15 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (colorless) Source: Recombinant | Order |
![]() | Platinum II Hot-Start Green PCR Master MixFidelity: 1x (standard) Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 15 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (green) Source: Recombinant | Order |
![]() | Platinum Direct PCR Universal Master MixFidelity: 1x (standard) Product capabilities: Universal annealing, Inhibitor tolerance, GC-rich PCR, Multiplex PCR, Long-range PCR, Benchtop stability, Direct PCREnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤8 kb DNA synthesis speed: 20 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (green) Source: Recombinant | Order |
![]() | Platinum Taq DNA Polymerase High FidelityFidelity: 6x Product capabilities: Long-range PCR, Benchtop stabilityEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤15 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A / blunt Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Platinum PCR SuperMix High FidelityFidelity: 6x Product capabilities: Long-range PCR, Benchtop stabilityEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤15 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A / blunt Product format: 1.1X SuperMix Source: Recombinant | Order |
![]() | Platinum Taq DNA PolymeraseFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Platinum Taq Green Hot Start DNA PolymeraseFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (green) Source: Recombinant | Order |
![]() | Platinum Hot Start PCR Master MixFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (colorless) Source: Recombinant | Order |
![]() | Platinum Green Hot Start PCR Master MixFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (green) Source: Recombinant | Order |
![]() | AmpliTaq Gold 360 DNA PolymeraseFidelity: 1x (standard) Product capabilities: GC-rich PCR, Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Chemical Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | AmpliTaq Gold 360 Master MixFidelity: 1x (standard) Product capabilities: GC-rich PCR, Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Chemical Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (colorless) Source: Recombinant | Order |
![]() | AmpliTaq Gold DNA PolymeraseFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Hot start Hot-start type: Chemical Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Platinum Taq DNA Polymerase, DNA-freeFidelity: 1x (standard) Product capabilities: Benchtop stability, Uracil/dUTP tolerance, Compatibility with TaqMan probe, Certified low bacterial DNAEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | DreamTaq DNA PolymeraseFidelity: 1x (standard) Product capabilities: Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | DreamTaq Green DNA PolymeraseFidelity: 1x (standard) Product capabilities: Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (green) Source: Recombinant | Order |
![]() | DreamTaq PCR Master MixFidelity: 1x (standard) Product capabilities: Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (colorless) Source: Recombinant | Order |
![]() | DreamTaq Green PCR Master MixFidelity: 1x (standard) Product capabilities: Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: 2X master mix (green) Source: Recombinant | Order |
![]() | AmpliTaq DNA PolymeraseFidelity: 1x (standard) Product capabilities: Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | AmpliTaq 360 DNA PolymeraseFidelity: 1x (standard) Product capabilities: GC-rich PCR, Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | AccuPrime Pfx DNA PolymeraseFidelity: 26x Product capabilities: Long-range PCR, Benchtop stabilityEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤12 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: Blunt Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | AccuPrime Pfx SuperMixFidelity: 26x Product capabilities: Long-range PCR, Benchtop stabilityEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤12 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: Blunt Product format: 1.1X SuperMix Source: Recombinant | Order |
![]() | Platinum Multiplex PCR Master MixFidelity: 1x (standard) Product capabilities: Multiplex PCR, Benchtop stability, Uracil/dUTP toleranceEnzyme type: Hot start Hot-start type: Antibody Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Invitrogen Taq DNA polymeraseFidelity: 1x (standard) Product capabilities: Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Recombinant | Order |
![]() | Invitrogen Taq DNA polymerase, nativeFidelity: 1x (standard) Product capabilities: Uracil/dUTP tolerance, Compatibility with TaqMan probeEnzyme type: Standard Taq Hot-start type: n/a Amplification length: ≤5 kb DNA synthesis speed: 60 sec/kb Amplicon overhang: 3′ A Product format: Stand alone (colorless) Source: Native | Order |
Search Thermo Fisher Scientific