Workflow automation and consumables
Autosamplers, robotic automation options, and data integration software can streamline your workflow and increase throughput. You'll also need liquids, bottles, beads and other consumables to run your Attune flow cytometry experiments.
Choose your system, lasers, filters and accessories—fast
Autosamplers
Improve workflow efficiency by choosing the autosampler option that best fits your throughput and experiment requirements. Two models of autosamplers are available to deliver walkaway automation seamlessly integrated with your Attune Flow Cytometer for increased productivity.
“The Attune NxT autosampler allowed us to process multiple 96-well plates in a day, enabling high throughput experiments that could not be performed on other flow cytometers.”
James Hemphill
PhD, Senior Scientist
Kytopen
Here is what the Attune CytKick Autosampler and the CytKick Max Autosampler have to offer:
CytKick Autosampler
CytKick Max Autosampler
Throughput
42 min for 96-well plate using high-throughput mode 22 min for 96-well plate (Boost mode, using one rinse and one mix and full analysis of a 20 µL sample)
Carryover
<0.5% carryover for 100 μL, 200 μL, 500 μL, and 1,000 μL samples with one mix and one rinse in standard mode
<1.0% carryover for 12.5 μL and 25 μL samples
<0.5% carryover for 100 μL, 200 μL, 500 μL, and 1,000 μL samples with one mix and one rinse in standard mode
<1.0% carryover for 12.5 μL and 25 μL samples
<1.0% carryover for 500 μL and 1,000 μL samples in Boost mode with one mix and one rinse
Compatibility
96 standard- and deep-well plates
384 standard- and deep-well plates
96 standard- and deep-well plates; foil-covered option
384 standard- and deep-well plates
1.5 mL and 2 mL microcentrifuge tube rack (up to 24 per rack)
Customizable to accept additional plate types
Cooling
--- Passive cooling available for 96-well U-bottom plates and microcentrifuge tube racks
“The Attune NxT is a very well-built instrument, we are very happy with the Attune NxT. The automated Autosampler is a real plus when it is combined with the acoustic focusing because it allows you to count very diluted samples fast. It’s very precise and gives reliable event counting.”
Hervé Luche
PhD, Senior Director, CIPHE
the Centre for ImmunoPHEnomics, Marseille, France
Our autosamplers are compatible with many different plate formats, including 96-well, 384-well, and up to 2 mL deep-well plates. The autosamplers give you a range of flexibility. An intelligent probe design minimizes clogging and carryover (0.5%) and helps prevent damage to the instrument. Conversion between plates and tubes is easy with a one-click transition. And autocleaning is performed automatically upon instrument shutdown. 
Unlike other systems that use shaking for mixing, our autosamplers aspirate samples for mixing. Aspiration for mixing is a novel approach that both ensures homogeneity of the sample and is gentle on cells to help ensure cell viability. 
Consistent results across all wells and across plates
Configuring Attune Flow Cytometer automation with the Attune CytKick and CytKick Max Autosampler demonstrates consistent results regardless of whether the sample is delivered from a tube or from a plate.
Dot plots of CD8 R-PE fluorescence vs CD4 FITC fluorescence from same samples in tubes vs plates
Attune NxT Autosampler offers consistent results regardless of sampling method 
Lysed whole blood was stained for CD45, CD4, and CD8 markers, aliquoted into both a tube and a plate, and analyzed on an Attune NxT flow cytometer. Minimal variation was observed between the sampling modes.
To determine consistency across wells and plates, lysed human whole blood was labeled with fluorophore-conjugated monoclonal antibodies for CD45, CD3, CD4, and CD8 targets. The labeled cells were fixed, then added in equal amounts to each well of two 96-well plates, and acquired on a 4-laser Attune NxT Flow Cytometer with Autosampler. Both percent positive (left) and concentration (events/µL, right) were highly consistent across wells for all three identified populations.
Consistent percentage data across plates and wells
Consistency of percent positive across 192 Samples
Consistency of all 192 wells for analysis of percent positive of parent gate of the three identified populations. 
Consistent concentration data across plates and wells
Consistency of concentration across 192 Samples
Consistency of all 192 wells for analysis of concentration of the three identified populations. 
Robotic automation
Extend your unmanned runtime settings and scalability with robotic integration applications for the Attune Flow Cytometers. Our range of automation solutions include robotic plate taxiing, extended fluidics, temperature-stable plate storage, and software for operations. You can leverage both scheduling and integration to get the most from your solution.
Attune CytPix Flow Cytometer with Attune CytKick Max Autosampler and Orbitor RS2 Microplate Mover
Attune CytPix Flow Cytometer with Attune CytKick Max Autosampler and Orbitor RS2 Microplate Mover
The robotic arm offers active and passive protective safety features, demonstrated reliability, and flexible configuration options for arrangement and storage. Operation is managed by Thermo Scientific Momentum Scheduling Software, established with instrument drivers available for over 200 instruments. The dashboard facilitates dynamic scheduling for active prioritization, visualized progress, and plate tracing. Extended-run fluidics allow for up to 12 hours of unattended continuous runtime under specific run conditions.
Orbitor RS2 Microplate Mover
The Orbitor RS2 Microplate Mover can handle 20 standard microplates or 9 deep-well blocks and a self-scanning internal inventory to help ensure flexibility. To mitigate evaporation, the system can de-lid and re-lid plates as they are loaded, unloaded, and stored.
Orbitor RS microplate mover
Thermo Scientific Orbitor RS2 Microplate Mover
The Orbitor RS2 is a compact platemover with remarkable speed and accuracy for robotically automated flow cytometry. The system’s advanced bi-directional telescoping arm provides exceptional reach and precision, making it one of the most flexible movers in the market. The integrated barcode scanner permits sample tracking, barcode transmission, and inventory management, while the plate detection in the gripper helps to minimize labware handling errors and reduces the risk of lost samples.
Cytomat automated incubator
The Cytomat automated incubator uses orbital shaking for consistency and dependability. The synchronized dual magnetic drive system (located at the bottom and top) helps ensure consistent shaking amplitude across all microplates regardless of load, position or number of plates. An inductive wear-free driving mechanism has no particle emission or belts to change.
The system supports a wide range of applications with individual speed settings of each Tower Shaker from 100-1200 rpm with oscillating amplitude of 2 mm (3 mm option available).
Cytomat Automated Incubator
Cytomat automated incubator
The Cytomat incubator is flexible and easy to set up.  An active microplate clamp mechanism secures plate and lid separately to prevent abrasions on microplates during shaking. For optimal assay flexibility you can mix both Standard and Tower Shaker. 
External Fluid Supply (EFS)
Attune NxT external fluid supply
Attune NxT External Fluid Supply (EFS)
This 10L External Fluid Supply extends the period of continual operation. The EFS fits under the bench or on an adjacent cart connected to the Attune Flow Cytometer by umbilical cords. The EFS is actively monitored and the operator is notified of issues encountered. 
Data integration software
Dynamic scheduling in Momentum Workflow Scheduling Software allows users to successfully create multiple workflows.
The software’s compatibility drivers support over 200 instruments, which have proven success in numerous implementations and can speed your integration project. 
Momemtum Monitor
Momentum Workflow Scheduling Software 
The software includes scheduling features to handle multiple workflows and compatibility drivers to speed implementation. 
A key factor to choosing your automation solution is ensuring that the software is both performant and easy to use. The dashboard in the Momentum Integration Software is easy to learn and use, and helps users to actively prioritize and reprioritize runs, visualize progress and trace plates.
Consumables
Bottles
Filters
Catalog # Name Size Price (USD) Qty
100083194 Each
436.00
Kits
Catalog # Name Size Price (USD) Qty
A43038 Each
3,180.00
100022775 Each
2,035.00
100022776 Each
1,226.65
Online Exclusive
1,510.00
Save 283.35 (19%)
A50920 Each
1,278.00
Performance tracking beads
Catalog # Name Size Price (USD) Qty
4449754 Each
222.65
Online Exclusive
244.00
Save 21.35 (9%)
Solutions
Replacement parts
For Research Use Only. Not for use in diagnostic procedures.