1x1 image pixel for data collection
Nunc™ 50 mL Bioreactor Tube
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ 50 mL Bioreactor Tube
Nunc™ 50 mL Bioreactor Tube
Nunc™ 50 mL Bioreactor Tube
Nunc™ 50 mL Bioreactor Tube
Thermo Scientific™

Nunc™ 50 mL Bioreactor Tube

Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition.
Have Questions?
Catalog number 332260
Price (USD)
128.65
Online Exclusive
140.00
Save 11.35 (8%)
Case of 90
In stock
Add to cart
Recurring order eligible. Learn more »
Price (USD)
128.65
Online Exclusive
140.00
Save 11.35 (8%)
Case of 90
Add to cart
Ask our AI about this Product
Thermo Scientific™ Nunc™ Bioreactor Centrifuge Tubes are designed for high throughput protein expression screening and cell line development and optimization under suspension condition. Adjustable 4-level vented cap with a 0.2 μm filter membrane allows for controlled gas exchange and air circulation without contamination. Ergonomic grooved cap for effortless handling and a large writing area for ease of identification.
  • Ideal for screening, small volume high throughput analysis
  • Protein expression suspension culture volumes ranging from 10 mL to 25 mL
  • Convenient pack sizes
  • Four different sizes of ventilation (dial type) on the cap.
  • Tube: Polypropylene
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene
  • 2 μm PTFE filter membrane
  • Suitable for suspension culture
  • Sterile - SAL 10-6
  • Non-pyrogenic
  • Non-cytotoxic
  • Shelf life: 2 years
Specifications
Closure MaterialHDPE
Dnase FreeYes
For Use WithIdeal for screening, small volume high throughput analysis
Includes
  • Cap: HDPE
  • Filter: PTFE
  • Dial: Polystyrene 
  • 0.2 μm PTFE filter membrane
No. per Case90
No. per Pack3
RNase FreeYes
AutoclavableAutoclavable
Bottom ShapeConical
Capacity (Metric)Protein expression suspension culture volumes ranging from 10 mL to 25 mL
Closure TypeQuadruple-position Vent/Close Cap
DisposableSingle-use
MaterialPolypropylene
Product TypeConical Centrifuge Tube
SterilitySterile
Writing AreaYes
Unit SizeCase of 90
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection