Keratinocyte SFM (1X), without calcium chloride
Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Product Image
Gibco™

Keratinocyte SFM (1X), without calcium chloride

Keratinocyte SFM (serum-free medium) is optimized for the growth and maintenance of human keratinocytes and other types of epithelial cellsRead more
Have Questions?
Catalog NumberQuantity
37010022500 mL
Catalog number 37010022
Price (USD)
162.65
Online Exclusive
182.00
Save 19.35 (11%)
Each
In stock
Add to cart
Quantity:
500 mL
Recurring order eligible. Learn more »
Price (USD)
162.65
Online Exclusive
182.00
Save 19.35 (11%)
Each
Add to cart
Ask our AI about this Product
Keratinocyte SFM (serum-free medium) is optimized for the growth and maintenance of human keratinocytes and other types of epithelial cells without the need for a feeder layer of cells. The complete medium may be used to cultivate human epidermal keratinocytes for studies involving dermal substitutes, in vitro toxicology, and gene therapy. Keratinocyte SFM without calcium is available to allow researchers to adjust the calcium concentration to meet the demands of their specific application.

Keratinocyte SFM is a complete serum-free medium when supplemented with human recombinant epidermal growth factor (rEGF) and bovine pituitary extract (BPE) at the time of use. Both supplements, rEGF and BPE, are supplied in separate packaging with the basal medium.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEpithelial Cells
Concentration1 X
Product TypeKit
Quantity500 mL
Shelf Life12 Months
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
Sterilization MethodSterile-filtered
With AdditivesGlutamine, Phenol Red
Without AdditivesNo Calcium Chloride
Unit SizeEach
Contents & Storage
• 500 mL Keratinocyte-SFM Basal Medium; store in refrigerator (2–8°C) and protect from light
• 25 mg Bovine Pituitary Extract (BPE); store in freezer (-5 to -20°C) and protect from light
• 2.5 μg EGF, Human Recombinant; store in freezer (-5 to -20°C) and protect from light

Complete Medium: store in refrigerator (2–8°C) and protect from light

Have questions about this product? Ask our AI assisted search.
For how long can I store Keratinocyte-SFM at 4 degrees C?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The base Keratinocyte-SFM has a shelf life of 12 months when stored at 2 to 8 degrees C, protected from light. Keratinocyte-SFM Supplement has a shelf life of 18 months when stored at -20 to 5 degrees C. The complete media should not be stored for longer than 4 weeks at 4 degrees C

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Share catalog number, name or link

1x1 image pixel for data collection