Caspases, key regulators of apoptosis, play an important role in programmed cell death. Caspase enzymes are responsible for cleaving protein substrates and executing the dismantling of cellular components during apoptosis. Thermo Fisher Scientific offers reagents and kits for detecting caspase activity in real-time or as end-point assays. Real-time caspase activity assays are an easy-to-use, no-wash method that allow for the visualization and monitoring of caspase activity in live cells. End-point caspase activity assays offer a reliable and convenient method for quantifying caspase activity at a specific time point. Our assays enable researchers to gain valuable insights into the mechanisms underlying cell death processes.
What are caspases?
Caspases are a family of protease enzymes that are crucial mediators of programmed cell death or apoptosis. They are divided into two groups: initiator caspases (e.g., caspase-8 and -9) and executioner caspases (e.g., caspase-3 and -7). Initiator caspases activate executioner caspases, which then function to cleave structural proteins and activate other enzymes (1, 2). 
Caspase proteases are synthesized as inactive precursors, also known as procaspases. Activation of initiator caspases occurs by dimerization, induced proximity, or association with specific binding partners. Executioner caspases are then activated by these initiator caspases or other activated executioner caspases. These executioner caspases then cleave specific proteins to start the cascade of events involved in cell death (3).
Caspases are not only involved in apoptosis but have been demonstrated to be involved in non-cell death processes such as inflammation, dendrite trimming, and cell differentiation (4). Dysregulation of caspases has been shown to contribute to various pathological conditions and human disease such as tumorigenesis, autoimmunity, autoinflammation, cancer, and neurodegenerative disorders (5,6). 
Caspase assays
Caspase assays are designed to detect and quantify caspase activity within cells, providing valuable insights into the apoptotic process. The use of specific substrates allows researchers to measure the enzymatic activity of caspases evaluating apoptosis over time or at a specific time. Since cell death cascades are complex and dynamic, it is highly recommended that a multi-parametric approach is conducted for an accurate assessment of apoptosis.
Learn about alternative apoptosis assays
CellEvent Caspase-3/7 Green
CellEvent Caspase-3/7 RedCellEvent Caspase-3/7 Green and RedCellEvent Caspase-3/7 Green ReadyProbes Reagent
ApplicationNo-wash assay to monitor ongoing caspase-3/7 activity.
ReadoutFollowing interaction with active caspase-3/7 in apoptotic cells, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly.
Caspase target and functionCaspases-3 and -7 are downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.
Detection reagentDEVD conjugated to a nucleic acid binding dye 
Ex/Em (nm)502/530590/610502/530590/610502/530
Suggested filterFITCTexas RedFITCTexas RedFITC
Instrument platform
  • Fluorescence microscopy
  • HCS
  • Microplate Reader
  • Fluorescence microscopy
  • HCS
  • Flow cytometer
Sample compatibility
  • Live cells
  • Reagent is fixable
  • Live cells
  • Reagent is fixable
Formatvarious1 vial (lyophilized powder)*
or
5 vials (lyophilized powder)
2 vials (1 Green, 1 Red)Dropper bottle
See all CellEvent Caspase-3/7 Detection ReagentsR37111
*1 vial=100 assays using 96-well plate; **5 vials=500 assays using 96-well plate
Watch this video of CellEvent Caspase-3/7 Green detection reagent in action
Video Player is loading.
Current Time 0:00
Duration 0:15
Loaded: 0%
Stream Type LIVE
Remaining Time 0:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    TMRM and CellEvent Caspase-3/7 Green used to monitor apoptosis progression in HeLa cells treated with staurosporine. HeLa cells were loaded with 50 nM TMRM (red) followed by 5 µM CellEvent Caspase 3/7 Reagent (green). Cells were then treated with 0.5 µM staurosporine and images were acquired every 5 minutes over 7 hours on a Molecular Devices ImageXpress at 10x. Over the 7 hour time course, staurosporine induced a loss of mitochondrial membrane potential followed by activation of caspase 3/7 as evidenced by a decrease in TMRM signal (red) and an increase in caspase 3/7 signal (green), respectively.
    How do you use CellEvent Caspase-3/7?
    CellEvent Caspase-3/7 is easy to use. Simply prepare fresh CellEvent Caspase-3/7 staining solution, then add directly to your cells, incubate for 30–60 minutes, and then visualize your cells (Figure 1). Apoptotic cells will have activated caspase-3/7 resulting in bright green or red nuclei, while cells without activated caspase-3/7 (non-apoptotic cells) will have minimal fluorescent signal.
    cellevent-3-7-protocol-image
    Figure 1. Schematic of CellEvent Caspase-3/7 protocol.
    No-wash caspase-3/7 assay for real-time monitoring
    CellEvent Caspase-3/7 detection reagent is an important tool for the study of apoptosis. It is available in both green (Ex/Em: 502/530) and red (Ex/Em: 590/610) and can be visualized with a FITC or Texas Red filter (respectively) on any fluorescence microscope. The cell-permeant CellEvent reagent comprises the four–amino acid peptide DEVD—which contains the recognition site for caspases 3 and 7—conjugated to a nucleic acid–binding dye. Because the DEVD peptide inhibits the ability of the dye to bind to DNA, CellEvent Caspase-3/7 Green detection reagent is intrinsically nonfluorescent. In the presence of activated caspase 3/7, the dye is cleaved from the DEVD peptide and free to bind DNA, producing a bright fluorescent signal indicative of apoptosis (Figure 2). This caspase assay is highly specific for caspase 3/7 activation and, as expected, we observe nearly complete inhibition of the CellEvent Caspase-3/7 Green detection reagent signal in cells pretreated with a caspase 3/7 inhibitor (Figure 3).
    brightfield and fluorescent overlay of cells with nuclei stained with blue or green fluorescence
    Figure 2. No-wash apoptosis detection in live cells. HeLa cells were loaded with 7.5 μM CellEvent Caspase-3/7 Green detection reagent and treated with vehicle control (A) or 0.5 μM staurosporine (B) for 4 hr. Untreated cells were also stained with Hoechst 33342 (blue) to visualize the nuclei of negative cells. The fluorescence signal is overlaid with differential interference contrast (DIC). Cells treated with staurosporine show a significant increase in signal from CellEvent Caspase-3/7 Green detection reagent (green).
    Figure 3. Inhibition of caspase 3/7 activity with a caspase inhibitor. HeLa cells were treated with 0.5 μM staurosporine in the presence of 0–30 μM Caspase 3/7 Inhibitor 1 (EMD Chemicals) for 4 hr. Cells were then labeled with 5 μM CellEvent Caspase-3/7 Green detection reagent and Hoechst 33342 (blue) in complete medium. (A) In a dose-dependent manner, the Caspase 3/7 Inhibitor 1 decreased the signal from CellEvent Caspase-3/7 Green detection reagent in staurosporine-treated cells, expressed quantitatively as percent positive cells. (B) Visualization of staurosporine-induced caspase 3/7 activity (green); and (C) inhibition thereof is also presented. Images were acquired and analyzed on the Thermo Scientific Cellomics ArrayScan VTI platform.
    One important advantage of the CellEvent Caspase-3/7 assay is that no wash steps are required, thus preserving fragile apoptotic cells that are typically lost during these rinses. The loss of apoptotic cells during wash steps may lead to an underestimation of the extent of apoptosis in the sample, resulting in poor assay accuracy. Additionally, the fluorescent signal resulting from cleavage of CellEvent Caspase-3/7 detection reagent survives formaldehyde fixation and detergent permeabilization, providing the flexibility to perform endpoint assays and to probe for other proteins using immunocytochemical techniques. CellEvent Caspase-3/7 Red, can also be imaged easily alongside GFP expression in live cells with little to no cross talk (Figure 4).
    caspase-red-gfp-localization
    Figure 4.  Expression of GFP with Caspase Red shows minimal crosstalk.  A673 and U2OS cells were plated at 3,000 cells per well and transduced with cytosolic GFP BacMam and treated with DMSO or 2 µM camptothecin for 24 hours at 37°C. Cells were subsequently stained with CellEvent Caspase 3/7 Red for 1 hour at 37°C prior to imaging on an EVOS M7000 widefield fluorescence microscope.
     Image-iT LIVE Green Caspase-3 and -7 Detection KitImage-iT LIVE Red Caspase-3 and -7 Detection KitImage-iT LIVE Green Poly Caspase Detection KitImage-iT LIVE Red Poly Caspase Detection Kit
    ApplicationEnd point assay for the detection of active caspases.
    ReadoutFluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added.
    Caspase target and functionImage-iT LIVE Green and Red Caspase-3 and -7 detect caspases-3 and -7, downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.Image-iT LIVE Green and Red Poly Caspase reagent reacts with multiple caspases (poly-caspases) that bind to the common recognition sequence valine-alanine-aspartic acid (VAD).
    Detection reagentFAM-DEVD-FMKSR-DEVD-FMKFAM-VAD-FMKSR-VAD-FMK
    Ex/Em (nm)488/530550/595488/530550/595
    Suggested filtersFITCTRITCFITCTRITC
    Nuclear counterstains
    Hoechst 33342
    Propidium iodide
    Hoechst 33342
    SYTOX Green
    Hoechst 33342
    Propidium iodide
    Hoechst 33342
    SYTOX Green
    Instrument platform
    Fluorescence microscopy
    HCS
    Fluorescence microscopy
    HCS
    Sample compatibilityLive cells
    Reagent is fixable
    Live cells
    Reagent is fixable
    Format25 assays25 assays
    See all Image-iT LIVE Red and Green Caspase Detection Kits
    Caspase assays for end point detection
    The Image-iT LIVE Caspase Detection Kits employ a novel approach to detect active caspases that is based on a fluorescent inhibitor of caspases (FLICA) methodology, essentially an affinity label. The reagent associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. For the caspase-3 and -7 reagents, this recognition sequence is aspartic acid-glutamic acid- valine-aspartic acid (DEVD); for the poly caspases reagents, this recognition sequence is valine-alanine-aspartic acid (VAD) and for caspase-8 reagent this recognition sequence is leucine-glutamic acid-threonine-aspartic acid (LETD). A fluorescent dye is attached as a reporter. The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase that was present at the time the inhibitor was added (Figure 5). Each kit contains a cell-impermeant nucleic acid stain to monitor plasma membrane integrity.
    FLICA reagents have been used widely to study apoptosis using flow cytometry and microscopy (7, 8). However it should be noted that while cellular fluorescence from the reagent is strongly linked to caspase activity in apoptotic cells, its interaction with other cellular sites may contribute to signal intensity in non-apoptotic cells (9). Appropriate controls should be included in any experimental design.
    Image-iT-LIVE-Red-Poly-Caspase-Detection-Kit-615x147
    Figure 5. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit (I35101). Cells were treated with staurosporine to induce apoptosis, and FLICA reagent (SR-VAD-FMK, left) was added. Cells were then loaded with SYTOX Green dye (middle) and Hoechst 33342 (blue fluorescence, right) and analyzed by fluorescence microscopy. Apoptotic, non-apoptotic, and dead cells are all visible in the same field.
    CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit
    Vybrant FAM Poly Caspases Assay Kit
    Vybrant FAM Caspase-3 and -7 Assay Kit
    Vybrant FAM Caspase-8 Assay Kit
    ApplicationNo-wash assay to monitor ongoing caspase-3/7 activity.End point assay for the detection of caspase activity.
    ReadoutFollowing interaction with active caspase-3/7, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly.Fluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added.
    Caspase target and functionDetects caspase-3 and -7, downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.Reacts with multiple caspases (poly-caspases) that bind common recognition sequence valine-alanine-aspartic acid (VAD).Reacts with caspase-3 and -7 that bind common recognition sequence aspartic acid-glutamic acid-valine-aspartic acid (DEVD).Reacts caspase-8 that bind common recognition sequence leucine-glutamic acid-threonine-aspartic acid (LETD).
    Detection reagentDEVD conjugated to a nucleic acid binding dyeFAM-VAD-FMKFAM-DEVD-FMKFAM-LETD-FMK
    Ex/Em (nm)511/533495/529388/530488/530
    Laser (nm)488488
    Nuclear counterstainSYTOX AADvancedHoechst 33342
    Propidium iodide
    Sample compatibilityLive cellsLive cells
    Format20 assays
    100 assays
    25 assays
     See all CellEvent Caspase-3/7 Green Assay KitsSee all Vybrant FLICA Caspase Apoptosis Assay Kits
    Real-time, no-wash caspase-3/7 assay for flow cytometry
    The CellEvent Caspase-3/7 Green flow cytometry assay allows for the detection of activated caspase-3 and caspase-7 in apoptotic cells. Cell-permeant CellEvent Caspase-3/7 detection reagent consists of a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. During apoptosis, caspase-3 and caspase-7 proteins are activated and able to cleave the caspase 3/7 recognition sequence encoded in the DEVD peptide. Cleavage of the recognition sequence and binding of DNA by the reagent labels the apoptotic cells with a bright, fluorogenic signal. When used together with the appropriate SYTOX dead cell stain, apoptotic cells can be easily discriminated from live and necrotic cells (Figure 6).
    bivariant flow plot showing apoptosis in drug-treated cells (right) versus control cells (left)
    Figure 6. Caspase activity detection in Jurkat cells using the CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit on the Attune Flow Cytometer. Jurkat cells (T-cell leukemia, human) were treated with (A) DMSO or (B) 10 µM camptothecin for 3 hours before labeling with the CellEvent Caspase 3/7 Green Flow Cytometry kit. Stained samples were analyzed on the Attune Acoustic Focusing Cytometer equipped with a 488-nm laser, and fluorescence emission was collected using a 530/30 BP filter for CellEvent Caspase 3/7 Green Detection Reagent and a 690/50BP filter for SYTOX AADvanced stain, respectively. Note that the treated cells have a higher percentage of apoptotic cells (panel B) than the basal level of apoptosis seen in the control cells (panel A). A = apoptotic cells, V = viable cells, N = necrotic cells.
    End-point caspase assays for flow cytometry
    The Vybrant FAM Caspase Assay Kits, also known as the Vybrant FLICA Caspase Apoptosis Assay Kits, are easy to use and do not require the need for special buffers to lyse or permeabilize the cell membranes. Simply add the reagent to your cell culture media, let it incubate 1–4 hours, wash the cells, and analyze (Figure 7).
    schematic of FLICA mechanism of action
    Figure 7.Detecting active caspases in apoptotic cells using the Vybrant FAM Caspase Assay Kits. Unbound FLICA reagent diffuses out of the cell and is washed away. The green-fluorescent signal is a direct measure of the amount of active caspase that was present at the time the inhibitor was added.
    These assays employ the fluorescent inhibitor of caspases (FLICA) methodology. The FLICA reagent, a caspase inhibitor, binds covalently and irreversibly to the reactive cysteines of active caspases. This binding inhibits further enzymatic activity and makes this an end point assay for caspase activity (Figure 8). Vybrant FAM Caspase Assay Kits allow for the accurate detection of active caspases in living cells, are more reliable than annexin V, and simpler to use than TUNEL assays. 
    bivariant flow plot showing apoptosis in drug-treated cells stained with FLICA and PI
    Figure 8. Detection of active caspases in living cells by flow cytometry.
    Jurkat cells were either treated with 10 μM camptothecin for 4 hours at 37ºC and 5% CO2 and stained with the FLICA reagent for caspase-3 and -7 and propidium iodide, both from the Vybrant FAM Caspase-3 and -7 Assay Kit. Samples were analyzed on a flow cytometer with 488 nm excitation using 530 nm bandpass and 670 nm longpass emission filters.
    What are caspases?
    Caspases are a family of protease enzymes that are crucial mediators of programmed cell death or apoptosis. They are divided into two groups: initiator caspases (e.g., caspase-8 and -9) and executioner caspases (e.g., caspase-3 and -7). Initiator caspases activate executioner caspases, which then function to cleave structural proteins and activate other enzymes (1, 2). 
    Caspase proteases are synthesized as inactive precursors, also known as procaspases. Activation of initiator caspases occurs by dimerization, induced proximity, or association with specific binding partners. Executioner caspases are then activated by these initiator caspases or other activated executioner caspases. These executioner caspases then cleave specific proteins to start the cascade of events involved in cell death (3).
    Caspases are not only involved in apoptosis but have been demonstrated to be involved in non-cell death processes such as inflammation, dendrite trimming, and cell differentiation (4). Dysregulation of caspases has been shown to contribute to various pathological conditions and human disease such as tumorigenesis, autoimmunity, autoinflammation, cancer, and neurodegenerative disorders (5,6). 
    Caspase assays
    Caspase assays are designed to detect and quantify caspase activity within cells, providing valuable insights into the apoptotic process. The use of specific substrates allows researchers to measure the enzymatic activity of caspases evaluating apoptosis over time or at a specific time. Since cell death cascades are complex and dynamic, it is highly recommended that a multi-parametric approach is conducted for an accurate assessment of apoptosis.
    Learn about alternative apoptosis assays
    CellEvent Caspase-3/7 Green
    CellEvent Caspase-3/7 RedCellEvent Caspase-3/7 Green and RedCellEvent Caspase-3/7 Green ReadyProbes Reagent
    ApplicationNo-wash assay to monitor ongoing caspase-3/7 activity.
    ReadoutFollowing interaction with active caspase-3/7 in apoptotic cells, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly.
    Caspase target and functionCaspases-3 and -7 are downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.
    Detection reagentDEVD conjugated to a nucleic acid binding dye 
    Ex/Em (nm)502/530590/610502/530590/610502/530
    Suggested filterFITCTexas RedFITCTexas RedFITC
    Instrument platform
    • Fluorescence microscopy
    • HCS
    • Microplate Reader
    • Fluorescence microscopy
    • HCS
    • Flow cytometer
    Sample compatibility
    • Live cells
    • Reagent is fixable
    • Live cells
    • Reagent is fixable
    Formatvarious1 vial (lyophilized powder)*
    or
    5 vials (lyophilized powder)
    2 vials (1 Green, 1 Red)Dropper bottle
    See all CellEvent Caspase-3/7 Detection ReagentsR37111
    *1 vial=100 assays using 96-well plate; **5 vials=500 assays using 96-well plate
    Watch this video of CellEvent Caspase-3/7 Green detection reagent in action
    Video Player is loading.
    Current Time 0:00
    Duration 0:15
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:15
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      TMRM and CellEvent Caspase-3/7 Green used to monitor apoptosis progression in HeLa cells treated with staurosporine. HeLa cells were loaded with 50 nM TMRM (red) followed by 5 µM CellEvent Caspase 3/7 Reagent (green). Cells were then treated with 0.5 µM staurosporine and images were acquired every 5 minutes over 7 hours on a Molecular Devices ImageXpress at 10x. Over the 7 hour time course, staurosporine induced a loss of mitochondrial membrane potential followed by activation of caspase 3/7 as evidenced by a decrease in TMRM signal (red) and an increase in caspase 3/7 signal (green), respectively.
      How do you use CellEvent Caspase-3/7?
      CellEvent Caspase-3/7 is easy to use. Simply prepare fresh CellEvent Caspase-3/7 staining solution, then add directly to your cells, incubate for 30–60 minutes, and then visualize your cells (Figure 1). Apoptotic cells will have activated caspase-3/7 resulting in bright green or red nuclei, while cells without activated caspase-3/7 (non-apoptotic cells) will have minimal fluorescent signal.
      cellevent-3-7-protocol-image
      Figure 1. Schematic of CellEvent Caspase-3/7 protocol.
      No-wash caspase-3/7 assay for real-time monitoring
      CellEvent Caspase-3/7 detection reagent is an important tool for the study of apoptosis. It is available in both green (Ex/Em: 502/530) and red (Ex/Em: 590/610) and can be visualized with a FITC or Texas Red filter (respectively) on any fluorescence microscope. The cell-permeant CellEvent reagent comprises the four–amino acid peptide DEVD—which contains the recognition site for caspases 3 and 7—conjugated to a nucleic acid–binding dye. Because the DEVD peptide inhibits the ability of the dye to bind to DNA, CellEvent Caspase-3/7 Green detection reagent is intrinsically nonfluorescent. In the presence of activated caspase 3/7, the dye is cleaved from the DEVD peptide and free to bind DNA, producing a bright fluorescent signal indicative of apoptosis (Figure 2). This caspase assay is highly specific for caspase 3/7 activation and, as expected, we observe nearly complete inhibition of the CellEvent Caspase-3/7 Green detection reagent signal in cells pretreated with a caspase 3/7 inhibitor (Figure 3).
      brightfield and fluorescent overlay of cells with nuclei stained with blue or green fluorescence
      Figure 2. No-wash apoptosis detection in live cells. HeLa cells were loaded with 7.5 μM CellEvent Caspase-3/7 Green detection reagent and treated with vehicle control (A) or 0.5 μM staurosporine (B) for 4 hr. Untreated cells were also stained with Hoechst 33342 (blue) to visualize the nuclei of negative cells. The fluorescence signal is overlaid with differential interference contrast (DIC). Cells treated with staurosporine show a significant increase in signal from CellEvent Caspase-3/7 Green detection reagent (green).
      Figure 3. Inhibition of caspase 3/7 activity with a caspase inhibitor. HeLa cells were treated with 0.5 μM staurosporine in the presence of 0–30 μM Caspase 3/7 Inhibitor 1 (EMD Chemicals) for 4 hr. Cells were then labeled with 5 μM CellEvent Caspase-3/7 Green detection reagent and Hoechst 33342 (blue) in complete medium. (A) In a dose-dependent manner, the Caspase 3/7 Inhibitor 1 decreased the signal from CellEvent Caspase-3/7 Green detection reagent in staurosporine-treated cells, expressed quantitatively as percent positive cells. (B) Visualization of staurosporine-induced caspase 3/7 activity (green); and (C) inhibition thereof is also presented. Images were acquired and analyzed on the Thermo Scientific Cellomics ArrayScan VTI platform.
      One important advantage of the CellEvent Caspase-3/7 assay is that no wash steps are required, thus preserving fragile apoptotic cells that are typically lost during these rinses. The loss of apoptotic cells during wash steps may lead to an underestimation of the extent of apoptosis in the sample, resulting in poor assay accuracy. Additionally, the fluorescent signal resulting from cleavage of CellEvent Caspase-3/7 detection reagent survives formaldehyde fixation and detergent permeabilization, providing the flexibility to perform endpoint assays and to probe for other proteins using immunocytochemical techniques. CellEvent Caspase-3/7 Red, can also be imaged easily alongside GFP expression in live cells with little to no cross talk (Figure 4).
      caspase-red-gfp-localization
      Figure 4.  Expression of GFP with Caspase Red shows minimal crosstalk.  A673 and U2OS cells were plated at 3,000 cells per well and transduced with cytosolic GFP BacMam and treated with DMSO or 2 µM camptothecin for 24 hours at 37°C. Cells were subsequently stained with CellEvent Caspase 3/7 Red for 1 hour at 37°C prior to imaging on an EVOS M7000 widefield fluorescence microscope.
       Image-iT LIVE Green Caspase-3 and -7 Detection KitImage-iT LIVE Red Caspase-3 and -7 Detection KitImage-iT LIVE Green Poly Caspase Detection KitImage-iT LIVE Red Poly Caspase Detection Kit
      ApplicationEnd point assay for the detection of active caspases.
      ReadoutFluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added.
      Caspase target and functionImage-iT LIVE Green and Red Caspase-3 and -7 detect caspases-3 and -7, downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.Image-iT LIVE Green and Red Poly Caspase reagent reacts with multiple caspases (poly-caspases) that bind to the common recognition sequence valine-alanine-aspartic acid (VAD).
      Detection reagentFAM-DEVD-FMKSR-DEVD-FMKFAM-VAD-FMKSR-VAD-FMK
      Ex/Em (nm)488/530550/595488/530550/595
      Suggested filtersFITCTRITCFITCTRITC
      Nuclear counterstains
      Hoechst 33342
      Propidium iodide
      Hoechst 33342
      SYTOX Green
      Hoechst 33342
      Propidium iodide
      Hoechst 33342
      SYTOX Green
      Instrument platform
      Fluorescence microscopy
      HCS
      Fluorescence microscopy
      HCS
      Sample compatibilityLive cells
      Reagent is fixable
      Live cells
      Reagent is fixable
      Format25 assays25 assays
      See all Image-iT LIVE Red and Green Caspase Detection Kits
      Caspase assays for end point detection
      The Image-iT LIVE Caspase Detection Kits employ a novel approach to detect active caspases that is based on a fluorescent inhibitor of caspases (FLICA) methodology, essentially an affinity label. The reagent associates a fluoromethyl ketone (FMK) moiety, which can react covalently with a cysteine, with a caspase-specific amino acid sequence. For the caspase-3 and -7 reagents, this recognition sequence is aspartic acid-glutamic acid- valine-aspartic acid (DEVD); for the poly caspases reagents, this recognition sequence is valine-alanine-aspartic acid (VAD) and for caspase-8 reagent this recognition sequence is leucine-glutamic acid-threonine-aspartic acid (LETD). A fluorescent dye is attached as a reporter. The FLICA reagent is thought to interact with the enzymatic reactive center of an activated caspase via the recognition sequence, and then to attach covalently through the FMK moiety. The FLICA inhibitor is cell permeant and noncytotoxic. Unbound FLICA molecules diffuse out of the cell and are washed away; the remaining fluorescent signal is a direct measure of the amount of active caspase that was present at the time the inhibitor was added (Figure 5). Each kit contains a cell-impermeant nucleic acid stain to monitor plasma membrane integrity.
      FLICA reagents have been used widely to study apoptosis using flow cytometry and microscopy (7, 8). However it should be noted that while cellular fluorescence from the reagent is strongly linked to caspase activity in apoptotic cells, its interaction with other cellular sites may contribute to signal intensity in non-apoptotic cells (9). Appropriate controls should be included in any experimental design.
      Image-iT-LIVE-Red-Poly-Caspase-Detection-Kit-615x147
      Figure 5. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit. Live HeLa cells visualized using the Image-iT LIVE Red Poly Caspase Detection Kit (I35101). Cells were treated with staurosporine to induce apoptosis, and FLICA reagent (SR-VAD-FMK, left) was added. Cells were then loaded with SYTOX Green dye (middle) and Hoechst 33342 (blue fluorescence, right) and analyzed by fluorescence microscopy. Apoptotic, non-apoptotic, and dead cells are all visible in the same field.
      CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit
      Vybrant FAM Poly Caspases Assay Kit
      Vybrant FAM Caspase-3 and -7 Assay Kit
      Vybrant FAM Caspase-8 Assay Kit
      ApplicationNo-wash assay to monitor ongoing caspase-3/7 activity.End point assay for the detection of caspase activity.
      ReadoutFollowing interaction with active caspase-3/7, the fluorogenic, cell-permeant substrate is able to bind DNA and fluoresce brightly.Fluorescent inhibitor of caspases interacts with the caspase enzymatic site. Following a wash step, the remaining fluorescence signal is a direct measure of the amount of active caspase at the time the inhibitor was added.
      Caspase target and functionDetects caspase-3 and -7, downstream effector caspases that, when activated, cleave protein substrates and execute apoptosis.Reacts with multiple caspases (poly-caspases) that bind common recognition sequence valine-alanine-aspartic acid (VAD).Reacts with caspase-3 and -7 that bind common recognition sequence aspartic acid-glutamic acid-valine-aspartic acid (DEVD).Reacts caspase-8 that bind common recognition sequence leucine-glutamic acid-threonine-aspartic acid (LETD).
      Detection reagentDEVD conjugated to a nucleic acid binding dyeFAM-VAD-FMKFAM-DEVD-FMKFAM-LETD-FMK
      Ex/Em (nm)511/533495/529388/530488/530
      Laser (nm)488488
      Nuclear counterstainSYTOX AADvancedHoechst 33342
      Propidium iodide
      Sample compatibilityLive cellsLive cells
      Format20 assays
      100 assays
      25 assays
       See all CellEvent Caspase-3/7 Green Assay KitsSee all Vybrant FLICA Caspase Apoptosis Assay Kits
      Real-time, no-wash caspase-3/7 assay for flow cytometry
      The CellEvent Caspase-3/7 Green flow cytometry assay allows for the detection of activated caspase-3 and caspase-7 in apoptotic cells. Cell-permeant CellEvent Caspase-3/7 detection reagent consists of a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. During apoptosis, caspase-3 and caspase-7 proteins are activated and able to cleave the caspase 3/7 recognition sequence encoded in the DEVD peptide. Cleavage of the recognition sequence and binding of DNA by the reagent labels the apoptotic cells with a bright, fluorogenic signal. When used together with the appropriate SYTOX dead cell stain, apoptotic cells can be easily discriminated from live and necrotic cells (Figure 6).
      bivariant flow plot showing apoptosis in drug-treated cells (right) versus control cells (left)
      Figure 6. Caspase activity detection in Jurkat cells using the CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit on the Attune Flow Cytometer. Jurkat cells (T-cell leukemia, human) were treated with (A) DMSO or (B) 10 µM camptothecin for 3 hours before labeling with the CellEvent Caspase 3/7 Green Flow Cytometry kit. Stained samples were analyzed on the Attune Acoustic Focusing Cytometer equipped with a 488-nm laser, and fluorescence emission was collected using a 530/30 BP filter for CellEvent Caspase 3/7 Green Detection Reagent and a 690/50BP filter for SYTOX AADvanced stain, respectively. Note that the treated cells have a higher percentage of apoptotic cells (panel B) than the basal level of apoptosis seen in the control cells (panel A). A = apoptotic cells, V = viable cells, N = necrotic cells.
      End-point caspase assays for flow cytometry
      The Vybrant FAM Caspase Assay Kits, also known as the Vybrant FLICA Caspase Apoptosis Assay Kits, are easy to use and do not require the need for special buffers to lyse or permeabilize the cell membranes. Simply add the reagent to your cell culture media, let it incubate 1–4 hours, wash the cells, and analyze (Figure 7).
      schematic of FLICA mechanism of action
      Figure 7.Detecting active caspases in apoptotic cells using the Vybrant FAM Caspase Assay Kits. Unbound FLICA reagent diffuses out of the cell and is washed away. The green-fluorescent signal is a direct measure of the amount of active caspase that was present at the time the inhibitor was added.
      These assays employ the fluorescent inhibitor of caspases (FLICA) methodology. The FLICA reagent, a caspase inhibitor, binds covalently and irreversibly to the reactive cysteines of active caspases. This binding inhibits further enzymatic activity and makes this an end point assay for caspase activity (Figure 8). Vybrant FAM Caspase Assay Kits allow for the accurate detection of active caspases in living cells, are more reliable than annexin V, and simpler to use than TUNEL assays. 
      bivariant flow plot showing apoptosis in drug-treated cells stained with FLICA and PI
      Figure 8. Detection of active caspases in living cells by flow cytometry.
      Jurkat cells were either treated with 10 μM camptothecin for 4 hours at 37ºC and 5% CO2 and stained with the FLICA reagent for caspase-3 and -7 and propidium iodide, both from the Vybrant FAM Caspase-3 and -7 Assay Kit. Samples were analyzed on a flow cytometer with 488 nm excitation using 530 nm bandpass and 670 nm longpass emission filters.
      Ordering information
      Catalog # Name Size Price (USD) Qty
      C10430 CellEvent™ Caspase-3/7 Red Detection Reagent (lyophilized), 1 Vial Each
      285.65
      Online Exclusive
      294.00
      Save 8.35 (3%)
      C10431 CellEvent™ Caspase-3/7 Red Detection Reagent (lyophilized), 5 Vials Each
      821.65
      Online Exclusive
      850.00
      Save 28.35 (3%)
      C10432 CellEvent™ Caspase-3/7 Green Detection Reagent (lyophilized), 1 Vial Each
      285.65
      Online Exclusive
      294.00
      Save 8.35 (3%)
      C10433 CellEvent™ Caspase-3/7 Green Detection Reagent (lyophilized), 5 Vials Each
      818.65
      Online Exclusive
      850.00
      Save 31.35 (4%)
      C10434 CellEvent™ Caspase-3/7 Variety pack (lyophilized), 2 Vials (1 Green, 1 Red) Each
      570.65
      Online Exclusive
      588.00
      Save 17.35 (3%)
      C10423 CellEvent™ Caspase-3/7 Green Detection Reagent, 1 x 100 μL Vial Each
      788.65
      Online Exclusive
      830.00
      Save 41.35 (5%)
      C10723 CellEvent™ Caspase-3/7 Green Detection Reagent, 1 x 25 μL Vial Each
      293.65
      Online Exclusive
      302.00
      Save 8.35 (3%)
      R37111 CellEvent™ Caspase-3/7 Green ReadyProbes™ Reagent Each
      341.65
      Online Exclusive
      369.00
      Save 27.35 (7%)
      Related categories
      decorative image
      decorative image
      decorative image
      decorative image
      decorative image
      decorative image
      Tools and handbooks
      Screen-Shot-2024-02-08-at-13.29.24
      Fluorescence SpectraViewer
      Supports all levels of experimental complexity, use the tool to compare excitation and emission spectra of fluorophores and reagents.
      Access tool
      decorative image
      Stain-iT Cell Staining Simulator
      Visualize staining your cell without wasting your reagents, antibodies, or time.
      Try now
      Learn how
       decorative image
      Flow cytometry protocols handbook
      Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
      Request the handbook
      decorative image
      Imaging protocol handbook
      Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies. 
      Request the handbook
      Support
      Cell Analysis Support Center
      Find technical information, tips and tricks, and answers to everyday problems.
      Stylesheet for Classic Wide Template adjustments
      For Research Use Only. Not for use in diagnostic procedures.