Product image of trypsin
Detach adherent cells and tissues with Gibco cell dissociation reagents
To acquire a suitable number of cells for research, a group of cells must first be disaggregated from a larger culture. Enter: cell dissociation. Gibco trypsin and alternative cell dissociation products are ideal for use with tissues and cell monolayers. Available in a variety of formats, both enzymatic and chemical, to meet the varied needs of researchers performing adherent cell culture. Explore the selection guide below to choose the correct cell dissociation reagent for your cell culture type.
Explore dissociation protocols
Choose the best cell dissociation reagent for your cell culture type
Cell culture typeWash solutionDissociation methodRecommended productFormatContains phenol red
Most continuous and strongly adherent cell linesHBSS or PBS w/o calcium and magnesiumTrypsin 0.25% in balanced salt solution w/o calcium and magnesium0.25% Trypsin (1X)LiquidYes
2.5% Trypsin (10X)LiquidNo
Trypsin (1:250)PowderNo
Many cells at early passageD-PBSTrypLE reagentTrypLE ExpressLiquidYes
Continuous cell lines where cell surface protein integrity is importantHBSS or PBS w/o calcium and magnesiumTrypsin 0.05% in 0.53 mM EDTA0.05% Trypsin 0.53 mM EDTALiquidYes
0.05% Trypsin 0.53 mM EDTA (10X)LiquidNo
TrypLE reagentTrypLE ExpressLiquidYes
Weakly adherent epithelial cellsHBSS or PBS w/o calcium and magnesiumEDTA, glycerol in sodium citrateCell Dissociation Buffer, Hanks'-basedLiquidNo
Cell Dissociation Buffer, PBS-basedLiquidNo
Transformed fibroblasts
HBSS or PBS w/o calcium and magnesiumTrypLE reagentTrypLE Express (1X)LiquidYes
Primary cells where cell surface protein integrity is importantHBSS or PBS w/o calcium and magnesiumTrypLE reagentTrypLE Express (1X)LiquidYes
Strongly adherent early-passage cell linesHBSS or PBS w/o calcium and magnesiumTrypsin 0.25% in 1 mM EDTA0.25% Trypsin 1 mM EDTALiquidYes
Trypsin Powder (1:250) PowderNo
Dispase 0.6–2.4 units/mL in PBSDispase IIPowder, LyophilizedNo
Epithelial cells0.5–1 mM EDTA0.5–1 mM EDTAVersene solution 1:5,000 (0.53 mM EDTA in PBS)LiquidNo
Dispase 0.6–2.4 units/mL in PBSDispase IIPowder, LyophilizedNo
Strongly adherent, epithelial and some tumor cell lines0.5–1 mM EDTA0.25% with 1 mM EDTA0.25% Trypsin 1 mM EDTALiquidYes
Versene solution 1:5,000 (0.53 mM EDTA in PBS)LiquidNo
Trypsin Powder (1:250) PowderNo
Dispase 0.6–2.4 units/mL in PBS w/o calcium and magnesiumDispase IIPowder, LyophilizedNo
Thick cultures, multiple layers1 mM EDTATrypsin 0.25%0.25% Trypsin 1 mM EDTALiquidYes
Trypsin Powder (1:250)PowderNo
Dense cultures that are collagen rich1 mM EDTACollagenase 200 units/mL, 1 mM EDTA in balanced salt solution w/o calcium and magnesiumCollagenase Type IPowder, LyophilizedNo
Collagenase Type IIPowder, LyophilizedNo
Collagenase Type IVPowder, LyophilizedNo
All adherent cell culturesHBSS or PBS w/o calcium and magnesiumPhysical/mechanicalCell scrapers
Trypsin and cell trypsinization
Trypsin, a proteolytic enzyme, is the standard way to detach adherent cell cultures and monolayers. This globular, pancreatic protease cleaves at the C-terminal side of lysine and arginine, breaking down vessel-adhering proteins and allowing easy resuspension during cell harvesting.
Thermo Fisher Scientific offers porcine-derived trypsin, EDTA-enhanced trypsin, as well as formulations of the TrypLE enzyme, a gentle, trypsin-like enzyme. To avoid unplanned protein degradation, organic trypsin must be inactivated with trypsin inhibitors. In contrast, synthetic trypsin, such as Gibco TrypLE products, are deactivated via dilution, eliminating the need for additional solutions.
NOTE: If you have fetal bovine serum (FBS) in your cell culture media, the culture will need to be washed with a balanced salt solution before trypsinization to ensure trypsin functionality.
Product image of trypsin
Trypsin-EDTA (0.25%), phenol red: a medium clear bottle with white and green labeling
Trypsin EDTA  
EDTA, a chelating agent, improves trypsin’s ability to detach adherent cells. Binding calcium and magnesium into a hexadentate, EDTA weakens cell-cell adhesion and enhances trypsin’s access to hydrolysis-targeted peptide bonds.
Gibco Trypsin-EDTA is made from irradiated porcine-derived protease with e-beam verification as well as tested for PPV, mycoplasma, and PCV 1/2 contamination.
Help enable gentle cell dissociation with TrypLE reagents
Gibco TrypLE reagents are highly purified, recombinant cell-dissociation enzymes that replace porcine trypsin. These reagents are ideal for dissociating attachment-dependent cell lines in both serum-containing and serum-free conditions, and can be directly substituted for trypsin without protocol changes. TrypLE reagents are gentle on cells, stable at room temperature, and animal origin–free.
Learn more about TrypLE reagents
 TrypsinTrypsin-EDTATrypLE ExpressTrypLE Select
CharacteristicsTraditional standard
Trypsin with enhanced cleavageGentler, stabler, and cost comparableGentler, stabler, animal and human-component free
Use caseGeneral purposeGeneral purposeGeneral purposeBioproduction/industrial application
Origin sourcePorcine pancreasPorcine pancreasFungus (animal origin-free)Fungus (animal origin-free)
Manufacturing standardscGMP manufacturingcGMP manufacturingcGMP manufacturingcGMP, manufactured on dedicated animal origin-free machinery
Storage temperature–5 to –20° C–5 to –20° CRoom temperatureRoom temperature
Higher cell viability  
Inactivation methodTrypsin inhibitorsTrypsin inhibitorsInhibition by dilutionInhibition by dilution
Trypsin inhibitors
Trypsin inhibitors destructively alter trypsin, nullifying the proteolytic enzyme’s ability to bind with, and so digest, proteins. Used post dissociation, trypsin inhibitors protect your cells against further protein degradation by trypsin.
NOTE: Soybean trypsin inhibitors also bind chymotrypsin, albeit to a lesser degree.
Trypsin inhibitors comparison chart
 Soybean trypsin inhibitor (powder)Defined trypsin inhibitorTrypsin neutralizer solution
Animal origin-free  
Calcium and magnesium free  
Sterile 
With 0.5% newborn bovine serum  
Catalog # Name Size Price (USD) Qty
R002100 Trypsin Neutralizer Solution Each
38.65
Online Exclusive
41.28
Save 2.63 (6%)
17075029 Soybean Trypsin Inhibitor, powder Each
223.00
R007100 Defined Trypsin Inhibitor Each
63.50
Collagenase
Relatively gentle, collagenase works by digesting connective tissue’s native triple-helical collagen fibrils. These tissues are often found in skin, tendon, blood vessels, and bone—making collagenase disaggregation suitable for human tumor, mouse kidney, human brain, and epithelium cultures.
Visit the Technical Reference Library to view protocols for collagenase dissociation of cells from primary tissue
Collagenase type IV product photo
Choose the right collagenase for your cell type
CollagenaseTissue/Cell type
Type IEpithelial, adrenal, lung, fat
Type IIHeart, thyroid, salivary, liver, bone, cartilage
Type IVIslet (insulin receptor sites)
Catalog # Name Size Price (USD) Qty
17101015 Collagenase, Type II, powder Each
365.65
Online Exclusive
383.00
Save 17.35 (5%)
17100017 Collagenase, Type I, powder, 1 g Each
370.65
Online Exclusive
387.00
Save 16.35 (4%)
17018029 Collagenase, Type I, powder, 500 mg Each
243.65
Online Exclusive
254.00
Save 10.35 (4%)
17104019 Collagenase, Type IV, powder Each
387.00
Dispase
Dispase, or neutral protease, is a Bacillus polymyxa–produced metalloenzyme. Rapid and gentle, dispase can be used to harvest and transfer normal diploid cells and cell lines. Dispase effectively separates cell clumps and splits cells from intact tissue with little impact to membrane integrity or viability.
Dispase II, powder in a small, white bottle with white and green labeling
Catalog # Name Size Price (USD) Qty
17105041 Dispase II, powder Each
909.65
Online Exclusive
956.00
Save 46.35 (5%)
Non-enzymatic cell dissociation
Versene solution
Versene solution, an EDTA solution in PBS, dissociates cells gently and non-enzymatically. This chelating agent detaches some cell types (such as epithelial cells) from vessel surfaces for suspension. It can also be used as a wash prior to trypsinization.
For further guidance on Versene solution applications, please see the cell dissociation selection guide by culture type.
Versene solution in a small, clear bottle with white and green labeling
Catalog # Name Size Price (USD) Qty
A4239101 CTS™ Versene™ Solution Each
43.68
15040066 Versene Solution Each
17.31
Non-enzymatic cell dissociation buffers
Recommended for weakly adherent cells, such as epithelial cells, Gibco cell dissociation buffers dissociate gently and maintain surface protein integrity. Applications include cells involved in ligand binding, flow cytometry, and immunohistochemistry research.
Please consult the enzyme-free cell dissociation buffer protocol for more information.
Try the Gibco MediaSelect Tool
Easily find a media solution suited to your specific application. Use this interactive tool to select classical basal, protein expression, or viral vector media.
Your selections will allow you to choose from a catalog menu of media – or customize your own formulation to fit your experimental setup and budget. Where available, you can compare media formulations side-by-side.
Find your media with the Gibco MediaSelect Tool
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Gibco Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments