Thermo Fisher Scientific is one of the few primary manufacturers of nucleotides in the industry. Our nucleotides—including dNTPs, NTPs, and modified nucleotides—are synthesized from high-quality raw materials in state-of-the-art production facilities and available in a variety of formats, formulations, and volumes for convenience and flexibility. Review our offerings below for your applications like PCR, cDNA synthesis, oligo synthesis and labeling, in vitro transcription, and many more.
For molecular assay developers and life science researchers looking for reproducible and reliable performance of dNTPs and NTPs, learn more about ISO 13485 quality standards that help to ensure batch-to-batch consistency from research to commercialization and inquire how we can support custom formulations for your commercial needs.
Need custom or bulk formulations?
Benefits of dNTPs, NTPs, and modified nucleotides
  • High purity (>99%) according to HPLC (Figure 1)
  • Free from DNase, RNase, and inhibitors of qPCR, PCR, and RT
  • Stable for three years at –20°C
  • Available in custom formulations and for commercial supply
Choose from a range of dNTP and NTP formulations
For deoxyribonucleotide triphosphate (dNTPs), choose from our catalog of individual products (dATP, dCTP, dGTP, dTTP, and dUTP). dNTPs are available in solutions of each individual species or as a mix of all four species. Custom formulations are also available for specific requirements.
All ribonucleotide triphosphate (NTPs) formulations are designed for convenience and flexibility. Individual nucleotides (ATP, CTP, GTP, and UTP) are supplied in concentrations up to 200 mM in Tris or sodium salt. Custom formulations and nucleotide mixes are also available.
Specifications for dNTPs and NTPs
Table 1. Specification for dNTPs and NTPs
 dNTPsNTPs
 dNTPsNTPs
Purity*>99%>99%
Presence of contaminating nucleotides, nucleosides and PCR inhibitors*None detectedNone detected
DNase, RNase, endo- and exodeoxyribonuclease**None detectedNone detected
Presence of human and E.coli DNA***NegativeNegative
Stability
  • 3 years at –20°C
  • >100 freeze-thaw cycles
  • 3 years at –20°C
  • >100 freeze-thaw cycles
Applications
  • qPCR, RT-qPCR
  • PCR, RT-PCR, cDNA synthesis
  • High-fidelity and long-range PCR
  • Isothermal amplification
  • DNA labeling
  • Cloning
  • Sanger sequencing and next-generation sequencing (NGS)
  • In vitro transcription
  • mRNA synthesis
  • siRNA synthesis
  • RNA amplification
  • Ligation
  • Phosphorylation
 Order nowOrder now
* Measured by HPLC (high performance liquid chromatography)
** Confirmed by incubation of labeled DNA or RNA target with NTPs or dNTPs
*** qPCR test for Alu repeats or 23S rRNA gene
† Stability studies assessed pH, HPLC profiles, and PCR performance.
Quality assurance for NTPs and dNTPs
Highly sensitive diagnostic assays and therapeutic regulatory standards require the highest nucleotide quality and batch-to-batch consistency. Our ISO 13485 and ISO 9001 certified dNTPs have a three year shelf life and are produced under stringent, multi-step QC and manufacturing standards, minimizing inconsistencies and errors in the final product. Manufactured with dedicated equipment in a zero-waste site, each production lot is assayed by the most stringent criteria regarding purity and functionality. For scalable, bulk, and flexible packaging, connect with our expert team for custom solutions.
Purity testing of nucleotide solutions
Purity testing assesses production lots for the desired species via HPLC (>99%) and monitors for the presence of nucleosidic byproducts, residual chemicals, or macromolecule contaminants that occur during production and purification processes. We use a variety of methods to assay for overall purity including HPLC, sensitive colorimetric assays, or performance in PCR (Table 5).
Table 5. Purity assessment of nucleotide solutions.
SpeciesDetection method
Nucleosidic contaminantHPLC
PyrophosphateMalachite green phosphate assay
Macromolecule contaminantsqPCR assay 
Triphosphate purity
We manufacture nucleotides with greater than 99% triphosphate purity as assessed by HPLC chromatography (Figure 1).
Graph showing the exceptional purity of NTPs
Figure 1. Exceptional purity of NTPs. HPLC analysis shows >99% triphosphate purity with negligible diphosphate and undetectable mono and tetraphosphate forms.
Nucleosidic contaminants
Trace amount of nucleosidic contaminants can significantly affect PCR performance. These include:
  • Mono- and diphosphate forms, which can decrease nucleotide incorporation
  • Di-deoxy base forms, which can terminate the amplification reaction
  • Modified nucleotides, which can decrease the sensitivity of an assay
Inorganic species
Residual chemicals used during nucleotide production or purification can interfere with PCR and are often termed “PCR inhibitors”. Pyrophosphate, a residual contaminant that can occur during production, can inhibit real-time PCR at concentrations of 0.1–0.3 mM, by interfering with fluorescence detection [1]. The presence of inorganic pyrophosphate is easily monitored using the Malachite Green Phosphate Assay which is a colorimetric method that detects formation of a complex between Malachite Green, molybdate, and free orthophosphate.
Macromolecule contaminants
The presence of macromolecules can cause false test results. These include:
  • DNase and RNase contaminants can affect cDNA synthesis resulting in false-negative results (Figure 2)
  • Nickase and protease activity can compromise the amplification template
  • Contaminating DNA from human and bacterial origins can cause false-positive results
Gel electrophoresis evaluation of ribonuclease-free Thermo Scientific dNTPs
Figure 2. Evaluation of ribonuclease-free Thermo Scientific dNTPs. Three dNTP formulations (all at 1 mM concentration) were incubated with 1 μg of RNA transcript (1 kb and 0.2 kb) for 4 hours at 37°C in 20 μL volume. The dNTP preparations are determined to be RNase free if there is no visible trace of RNA transcript degradation. M: Thermo Scientific RiboRuler Low Range RNA Ladder, ready-to-use. K1: Control RNA transcript, 1 kb long. K2: Control RNA transcript, 0.2 kb long. A:With dNTP mix, 2 mM each. B:With dNTP mix, 10 mM each. C:With dNTP set.
Functional testing for nucleotide quality control
To complement analytical tests for quality control, each lot of Thermo Scientific nucleotides is also analyzed using a variety of functional tests including RT-qPCR and in vitro transcription.
dNTPs
Samples of each lot are tested by RT-qPCR (Figure 3). The assay is performed with low template concentration, which allows detection of even minute variations in dNTP performance.
Amplification plot showing the high quality of dNTPs
Figure 3. High quality of dNTPs helps ensure consistent Ct values in qPCR, especially with low template concentrations. dNTPs were tested for detection of the GAPDH gene in two-step RT-qPCR using different amounts of RNA transcript (107 to 103 copies) in reverse transcription reactions followed by amplification with hot-start Taq DNA polymerase.
NTPs
Samples of each lot are tested by in vitro transcription (Figure 4).
Agarose gel showing NTP performance in high-yield in vitro transcription
Figure 4. NTP performance in high-yield in vitro transcription. RNA transcripts were generated using Thermo Scientific NTPs and NTPs from two other vendors in high-yield in vitro transcription reactions containing 1 μg of DNA template. M:Thermo Scientific RiboRuler High Range RNA Ladder, ready-to-use. 1, 4, 7: 0.5 kb RNA transcripts. 2, 5, 8: 1 kb RNA transcripts. 3, 6, 9: 6 kb RNA transcripts.
Nucleotide ordering information
Do you need Invitrogen dNTPs or NTPs for an existing protocol? Please see our Invitrogen Nucleotides Collection to find the products you need.
Thermo Scientific dNTPs
Catalog # Name Size Price (USD) Qty
R0141 dATP Solution (100 mM) Each
92.50
R0151 dCTP Solution (100 mM) Each
75.65
Online Exclusive
92.50
Save 16.85 (18%)
R0161 dGTP Solution (100 mM) Each
82.65
Online Exclusive
92.50
Save 9.85 (11%)
R1191 dITP Solution (100 mM) Each
89.65
Online Exclusive
101.00
Save 11.35 (11%)
R0191 dNTP Mix (10 mM each), 0.2 mL Each
38.65
Online Exclusive
47.00
Save 8.35 (18%)
R0192 dNTP Mix (10 mM each), 1 mL Each
130.65
Online Exclusive
158.00
Save 27.35 (17%)
R0241 dNTP Mix (2 mM each), 1 mL Each
46.65
Online Exclusive
53.00
Save 6.35 (12%)
R0193 dNTP Mix (10 mM each), 5 x 1 mL Each
510.65
Online Exclusive
596.00
Save 85.35 (14%)
R0242 dNTP Mix (2 mM each), 5 x 1 mL Each
180.65
Online Exclusive
194.00
Save 13.35 (7%)
R1121 dNTP Mix (25 mM each), 1 mL Each
252.65
Online Exclusive
299.00
Save 46.35 (16%)
R1122 dNTP Mix (25 mM each), 5 x 1 mL Each
990.65
Online Exclusive
1,154.00
Save 163.35 (14%)
R0181 dNTP Set, 100 mM Solutions, 4 x 0.25 mL Each
190.65
Online Exclusive
233.00
Save 42.35 (18%)
R0182 dNTP Set, 100 mM Solutions, 4 x 1 mL Each
648.65
Online Exclusive
720.00
Save 71.35 (10%)
R0186 dNTP Set, 100 mM Solutions, 4 x 5 mL Each
2,557.65
Online Exclusive
2,735.00
Save 177.35 (6%)
R0251 dNTP/dUTP Mix Each
81.65
Online Exclusive
85.25
Save 3.60 (4%)
R0171 dTTP Solution (100 mM) Each
80.65
Online Exclusive
93.00
Save 12.35 (13%)
R0133 dUTP Solution (100 mM) Each
90.50
R0194 dNTP Mix (10 mM each), 10 x 1 mL Each
732.65
Online Exclusive
786.00
Save 53.35 (7%)
Thermo Scientific NTPs
Catalog # Name Size Price (USD) Qty
R0441 ATP Solution (100 mM) Each
50.65
Online Exclusive
59.50
Save 8.85 (15%)
R0451 CTP Solution (100 mM) Each
52.65
Online Exclusive
59.00
Save 6.35 (11%)
R0481 NTP Set, 100 mM Solution Each
140.65
Online Exclusive
169.00
Save 28.35 (17%)
R0471 UTP Solution (100 mM) Each
49.65
Online Exclusive
59.00
Save 9.35 (16%)
R1441 ATP Solution, Tris buffered Each
54.65
Online Exclusive
61.00
Save 6.35 (10%)
R1451 CTP Solution, Tris buffered Each
56.65
Online Exclusive
60.25
Save 3.60 (6%)
R1461 GTP Solution, Tris buffered Each
53.65
Online Exclusive
61.00
Save 7.35 (12%)
R1471 UTP Solution, Tris buffered Each
54.65
Online Exclusive
60.25
Save 5.60 (9%)
Modified nucleotides
Custom service and flexible packaging
  • Format—any concentration, formulation, and volume; custom mixes and blends
  • Scale—microliters to liters, milligrams to kilograms
  • Packaging—labeling, finishing, and choice of vials and bottles
Connect with our expert team and build custom solutions for your commercial needs
Need higher grade nucleotides?
We also offer TheraPure NTPs and modified nucleotides for nucleic acid therapeutics development. These high-quality nucleotides are produced with strictly controlled manufacturing processes and well-defined impurity profiles.
Contact us to learn more at NATxOEM@thermofisher.com.
References
Support
  • Contact us
    Email or call our technical application scientists for additional questions about dNTPs, NTPs, and modified nucleotides.
  • OEM and custom solutions
    Contact us for tailor-made performance tested products with strict quality standard to meet your molecular assay requirements.
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.