Production Tanks and Drums
Durable and built for performance
Contain and store bioproduction fluids in these durable, all-purpose vessels. These production tanks are economical alternatives to stainless-steel tanks. Designed for use in closed and open systems, they are an excellent choice for reagent storage, aseptic mixing/re-hydration, and dispensing. These vessels are also good solutions for waste management.
Product highlights
Decorative image of closed dome tank
Store, mix and dispense reagents in Thermo Scientific Nalgene Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 76 to 380 L, tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sealing.
Key features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Decorative image of rectangular tank
These Thermo Scientific Nalgene Rectangular tanks feature seamless construction for easing cleaning and are suitable for use with many organic chemicals.
Key features:
  • Versatility at lower cost than stainless-steel tanks
  • Impact and abrasion resistant
  • Polypropylene or heavy-duty LLDPE construction are available
  • Polypropylene tanks are available in sizes ranging from 8 – 114 L
  • LLDPE tanks are available in sizes ranging from 19 – 208 L
Tanks can be modified with a factory installed spigot
Decorative image of cylindrical tank
These Thermo Scientific Nalgene cylindrical tanks are durable, all-purpose vessels suitable for containing and storing bioproduction fluids, these tanks are an economical alternative to stainless-steel tanks.
Key features:
  • Linear low density polyethylene (LLDPE),  polypropylene (PP) and heavy duty LLDPE construction are available
  • Impact and abrasion resistant
  • Available in a range of sizes from 19 – 757 L
  • Sizes up to 378 L available with factory-installed spigots
Decorative image of cylindrical drums
Thermo Scientific cylindrical drums provide a qualified shipping system for BioProcess Containers (BPCs) as well as in-process liquid handling applications.
Key features:
  • Linear low density polyethylene (LLDPE),  construction are available
  • Available in 50, 100, 200, and 250 L sizes 
  • Top- or bottom drain options available
  • Support a range of BPCs and tank liner designs
  • Used for in-process applications such as harvesting and storage, and in-process liquid handling
  • Validated for transportation of sterile process liquids    
Decorative illustration of tank liners
Designed specifically to fit Nalgene and Thermo Scientific cylindrical tanks and drums, these tank liners feature an open-bag, flat-bottom design to enhance your mixing capabilities and to maximize the utility of your carboys and tanks.
Key features:
  • Removes the need for tank cleaning and reduces the cycle times
  • Film availability: Aegis5-14, CX3-9, ASI 26, and Nalgene Coex polyethylene film.
  • Sizes ranging from 19 – 787 L
  • Gamma-irradiated
Decorative image of closed dome tank
Store, mix and dispense reagents in Thermo Scientific Nalgene Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 76 to 380 L, tanks feature a 150 mm neck opening fitted with a gasketed closure for secure sealing.
Key features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Decorative image of rectangular tank
These Thermo Scientific Nalgene Rectangular tanks feature seamless construction for easing cleaning and are suitable for use with many organic chemicals.
Key features:
  • Versatility at lower cost than stainless-steel tanks
  • Impact and abrasion resistant
  • Polypropylene or heavy-duty LLDPE construction are available
  • Polypropylene tanks are available in sizes ranging from 8 – 114 L
  • LLDPE tanks are available in sizes ranging from 19 – 208 L
Tanks can be modified with a factory installed spigot
Decorative image of cylindrical tank
These Thermo Scientific Nalgene cylindrical tanks are durable, all-purpose vessels suitable for containing and storing bioproduction fluids, these tanks are an economical alternative to stainless-steel tanks.
Key features:
  • Linear low density polyethylene (LLDPE),  polypropylene (PP) and heavy duty LLDPE construction are available
  • Impact and abrasion resistant
  • Available in a range of sizes from 19 – 757 L
  • Sizes up to 378 L available with factory-installed spigots
Decorative image of cylindrical drums
Thermo Scientific cylindrical drums provide a qualified shipping system for BioProcess Containers (BPCs) as well as in-process liquid handling applications.
Key features:
  • Linear low density polyethylene (LLDPE),  construction are available
  • Available in 50, 100, 200, and 250 L sizes 
  • Top- or bottom drain options available
  • Support a range of BPCs and tank liner designs
  • Used for in-process applications such as harvesting and storage, and in-process liquid handling
  • Validated for transportation of sterile process liquids    
Decorative illustration of tank liners
Designed specifically to fit Nalgene and Thermo Scientific cylindrical tanks and drums, these tank liners feature an open-bag, flat-bottom design to enhance your mixing capabilities and to maximize the utility of your carboys and tanks.
Key features:
  • Removes the need for tank cleaning and reduces the cycle times
  • Film availability: Aegis5-14, CX3-9, ASI 26, and Nalgene Coex polyethylene film.
  • Sizes ranging from 19 – 787 L
  • Gamma-irradiated
Related products
Available in standard or custom configurations in a full range of sizes for a range of applications    
Available in standard or custom configurations in a full range of sizes for a range of applications    
Ordering information
Closed-dome tanks
Rectangular tanks
Cylindrical drums
Cylindrical drums
Tank liners
Catalog # Name Size Price (USD)
SH3064701 HyPerforma™ 50 to 500L Tank Liners, 50L
SH3064702 HyPerforma™ 50 to 500L Tank Liners, 100L
SH3064703 HyPerforma™ 50 to 500L Tank Liners, 200L
SH3064601 HyPerforma™ 50 to 500L Tank Liners, 50L Liner - Bottom drain, irradiated
SH3064602 HyPerforma™ 50 to 500L Tank Liners, 100L Liner - Bottom drain, irradiated
SH3064603 HyPerforma™ 50 to 500L Tank Liners, 200L Liner - Bottom drain, irradiated
SH3064704 HyPerforma™ 50 to 500L Tank Liners, 50L
SH3064705 HyPerforma™ 50 to 500L Tank Liners, 100L
SH3064706 HyPerforma™ 50 to 500L Tank Liners, 300L
SH3064707 HyPerforma™ 50 to 500L Tank Liners, 500L Liner - Diameter: 31in. (79cm)
B100037I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film 200 L
B100038I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film 130 L
B100048I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film 50 L
B100049I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film, 340 L, 35 per case Each
B100050I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film, 400 L, 20 per case Each
B100051I Thermo Scientific™ 2D Tank Liner, ASI™ 26 film, 560 L, 20 per case Each
B100364I Thermo Scientific™ 3D Tank Liner, ASI™ 26 film 50 L
B100373I Thermo Scientific™ 3D Tank Liner, ASI™ 26 film 100 L
B100363I Thermo Scientific™ 3D Tank Liner, ASI™ 26 film 200 L
B101459I Thermo Scientific™ 3D Tank Liner, ASI™ 26 film, 300 L, 10 per case Each
B101460I Thermo Scientific™ 3D Tank Liner, ASI™ 26 film, 560 L, 10 per case Each
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: