Related Product Information
Several applications require the maintenance of human induced pluripotent stem cells (iPSCs) in the absence of a murine embryonic fibroblast (MEF) feeder layer. This protocol describes the culture of human iPSCs on matrix-coated plates in the presence of MEF-conditioned media (MEF-CM).
In this system, iPSCs are maintained at high density. At confluence, the cells reach 300,000–500,000 cells/cm2.
10828028,10565018,11140050,PHG0264,A1413301
Ordering Information
10828028,10565018,11140050,PHG0264,A1413301
- Dulbecco’s Modified Eagle Medium (D-MEM) (Cat. no. 10569-010)
- Fetal Bovine Serum (FBS), ESC-Qualified (Cat. no. 10439-024)
- D-MEM/F-12 (1X) with GlutaMAX™-I (Cat. no. 10565-018)
- KnockOut™ Serum Replacement (KSR) (Cat. no. 10828-028)
- MEM Non-Essential Amino Acids Solution, 10 mM (Cat. no. 11140-050)
- β-mercaptoethanol, 1000X (Cat. no. 21985-023)
- Gibco® Mouse Embryonic Fibroblasts (Irradiated) (Cat. no. S1520-100)
- Basic Fibroblast Growth Factor (bFGF) (Cat. no. PHG0264)
- GlutaMAX™-I (100X) (Cat. no. 35050-079)
- Collagenase Type IV (Cat. no. 17104-019) for enzymatic passaging or StemPro® EZPassage™ Tool (Cat. no. 23181-010) for mechanical passaging
- Cell Scraper (Falcon, Cat. no. 353085)
- Dulbecco’s PBS (DPBS) with Calcium and Magnesium (Cat. no. 14040-133)
- Dulbecco’s PBS (DPBS) without Calcium and Magnesium (Cat. no. 14190-144)
- Geltrex® LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix (Cat. no.
A1413301 or A1413302) or CELLstart™ CTS™ substrate (Cat. no. A1014201) - Attachment Factor (Cat. no. S006100)
- Stem Cell Tested 0.22 μm pore size sterile filters (Nalgene, Cat. no. 564-0020 or 156-4020)
- BD Falcon T175 TC-Flasks (Falcon, Cat. no. 353112)
- 37°C water bath
- Appropriate tissue culture plates and supplies
Note: As an alternative to D-MEM/F-12 (1X) with GlutaMAX™-I, KSR, and bFGF, you can use the KnockOut™
ESC/iPSC Media Kit (Cat. no. A1412901).
Preparing Media and Materials
10 μg/mL bFGF Solution (1000 μL)
- To prepare 1 mL of 10 μg/mL bFGF solution, aseptically mix the following components:
Component | Volume |
---|
bFGF | 10 μg |
DPBS without Calcium and Magnesium | 980 μL |
10% KSR | 10 μL |
2. Aliquot and store at –20°C for up to 6 months.
10 mg/mL Collagenase Type IV Solution
- 1. Add D-MEM/F-12 to Collagenase Type IV to make a 10 mg/mL solution. Gently vortex to suspend and
filter sterilize the solution. This solution can be aliquoted and frozen at –20°C until use. - The 10 mg/mL working solution can be used for 2 weeks if properly stored at 2–8°C (store in aliquots to avoid repeated warming).
MEF Medium (for 100 mL of complete medium)
- To prepare 100 mL of complete MEF medium, aseptically mix the following components:
Component | Volume |
---|
D-MEM | 89 mL |
FBS, ESC-Qualified | 10 mL |
MEM Non-Essential Amino Acids Solution, 10 mM | 1 mL |
β-mercaptoethanol, 1000X | 100 μL |
2. Complete MEF medium can be stored at 2–8°C for up to 1 week.
Pluripotent Stem Cell (PSC) Culture Medium (for 100 mL complete medium)
- To prepare 100 mL of complete PSC Culture Medium, aseptically mix the following components:
Component | Volume |
---|
D-MEM/F-12 | 79 mL |
KSR | 20 mL |
MEM Non-Essential Amino Acids Solution, 10 mM | 1 mL |
bFGF (10 μg/mL)* | 40 μL |
β-mercaptoethanol, 1000X | 100 μL |
*Add bFGF at the time of medium change (final concentration 4 ng/mL).
2. Complete MEF medium can be stored at 2–8°C for up to 4 weeks.
MEF-Conditioned Medium (MEF-CM)
- Cover the whole surface of each new culture vessel with Attachment Factor (AF) solution and incubate the vessels for 30 minutes at 37°C or for 1 hour at room temperature. For MEF-CM generation, a T-175 flask is recommended.
- Using sterile technique in a laminar flow culture hood, completely remove the AF solution from the culture vessel by aspiration just prior to use. Coated vessels may be used immediately or stored at room temperature for up to 24 hours.
Note: It is not necessary to wash the culture surface before adding cells or medium. - Plate 9.4 × 106 Mitomycin C-treated or irradiated MEFs in a T-175 flask coated with AF and containing
30 mL of MEF medium. - The following day, replace the MEF medium with 90 mL of PSC Culture Medium.
- Collect the PSC Culture Medium, now considered MEF-CM, from the flasks after 24 hours of conditioning for up to seven days in a row.
- Each day, filter sterilize the collected MEF-CM with a 0.22 μM filter. Filtered MEF-CM can be stored at
–20°C until use. - At the time of use, thaw the MEF-CM in a 37°C waterbath, and freshly supplement it with additional bFGF (20 ng/mL).
Coating Culture Vessels with Geltrex® LDEV-Free, hESC-Qualified Basement Membrane Matrix
- Thaw a 5-mL bottle of Geltrex® LDEV-Free hESC-Qualified Reduced Growth Factor Basement Membrane Matrix at 2–8°C overnight.
- Dilute the thawed Geltrex® solution 1:1 with cold sterile D-MEM/F-12 to prepare 1-mL aliquots in tubes
chilled on ice. These aliquots can be frozen at –20°C or used immediately.
Note: Aliquot volumes of 1:1 diluted Geltrex® solution may be adjusted according to your needs. - To create working stocks, dilute a Geltrex® aliquot 1:50 with cold D-MEM on ice, for a total dilution of 1:100.
Note: An optimal dilution of the Geltrex® solution may need to be determined for each cell line. Try various dilutions from 1:30 to 1:100. - Quickly cover the whole surface of each culture dish with the Geltrex® solution (refer to Table 1).
- Incubate the dishes in a 37°C, 5% CO2 incubator for 1 hour.
Note: Dishes can now be used or stored at 2–8°C for up to a week. Do not allow dishes to dry. - Aspirate the diluted Geltrex® solution from the culture dish and discard. You do not need to rinse off the
Geltrex® solution from the culture dish after removal. Cells can now be passaged directly into MEF-CM onto the Geltrex® matrix-coated culture dish.
Note: CELLstart™ CTS™ substrate may be substituted for Geltrex® hESC-Qualified Matrix (see the Appendix).
Table 1. Volume of Geltrex® hESC-Qualified Matrix Required
Culture Vessel | Surface Area (cm2) | Volume of Diluted Substrate (mL) |
---|
6-well plate | 10 cm2/well | 1.5 mL per well |
12-well plate | 4 cm2/well | 750 μL per well |
24-well plate | 2 cm2/well | 350 μL per well |
35-mm dish | 10 cm2 | 1.5 mL |
60-mm dish | 20 cm2 | 3.0 mL |
100-mm dish | 60 cm2 | 6.0 mL |
TOP
Thawing and Plating iPSCs
- Label the Geltrex® matrix-coated dish with the passage number from the vial, the date, and user initials.
- Remove the vial of iPSCs from liquid nitrogen storage using metal forceps.
Note: If the vial is exposed to ambient temperatures for more than 15 seconds between removal and thawing, transfer the vial into a container containing a small amount of liquid nitrogen. - Roll the vial between your gloved hands until the outside is free of frost. This should take ~10–15 seconds.
- Immerse the vial in a 37°C water bath without submerging the cap. Swirl the vial gently.
- When only an ice crystal remains, remove the vial from the water bath, spray the outside of the vial with
70% ethanol to sterilize, and place it in hood. - Pipet the thawed cells gently into a sterile 50-mL conical tube using a 5-mL sterile pipette.
- Slowly add 10 mL of MEF-CM drop-wise to cells in the 50-mL conical tube. While adding the medium,
gently move the tube back and forth to mix the iPSCs. This reduces osmotic shock to the cells. - Rinse the vial with 1 mL of MEF-CM and add to the 50-mL conical tube with cells.
- Transfer cell suspension into a 15-mL centrifuge tube. Centrifuge the cells at 200 × g for 5 minutes.
- Aspirate and discard the supernatant.
- Resuspend the cell pellet in sufficient volume of MEF-CM according to Table 2 by gently pipetting the cells up and down in the tube a few times.
- Aspirate the excess Geltrex® solution from the prepared dish and slowly add the thawed colonies onto the dish. Move the dish in several quick, short, back-and-forth and side-to-side motions to disperse cells across the surface the dish.
- Place dish gently into the 37°C, 5% CO2 incubator and incubate the cells overnight.
- The next day, remove the spent medium with debris using a sterile serological pipet and transfer it into a prepared Geltrex® matrix-coated dish. You can use this dish as a backup in case there is a problem with the main dish.
- Add fresh MEF-CM to each dish according to the volumes in Table 2. Place both plates gently into a 37°C, 5% CO2 incubator overnight.
- Examine cells under the microscope and replace spent medium daily from both plates. If feeding more than one plate, use a different pipette for each well to reduce the risk of contamination. Colonies may not be visible for up to a week.
Table 2. Volume of MEF-CM Required
Culture Vessel | Surface Area (cm2) | Volume (mL) |
---|
6-well plate | 10 cm2/well | 2.0 mL per well |
12-well plate | 4 cm2/well | 1.0 mL per well |
24-well plate | 2 cm2/well | 0.5 mL per well |
35-mm dish | 10 cm2 | 2.0 mL |
60-mm dish | 20 cm2 | 4.0 mL |
100-mm dish | 60 cm2 | 10.0 mL |
Figure 1. PSCs growing in MEF-CM on a Geltrex® matrix-coated dish
When to Split Cells
In general, split cells when one of the following occurs:
- iPSC colonies are becoming too dense or too large.
- Increased differentiation occurs.
Split Ratio
- The split ratio can vary, though it is generally between 1:2 and 1:4. Occasionally, cells will grow at a different rate and the split ratio will need to be adjusted. A general rule is to observe the last split ratio and adjust the ratio according to the appearance of the iPSC colonies.
- If the cells look healthy and colonies have enough space, split using the same ratio. If they are overly dense and crowding, increase the ratio. If the cells are sparse, decrease the ratio. Cells will need to be split every 4–10 days based upon appearance.
Enzymatic Passaging Using Collagenase
You may passage cells via the enzymatic method as described below, or mechanically as described in the
following section.
- Label a new Geltrex® matrix-coated dish with the cell line name, the new passage number, the date, the split ratio, and user initials. Return the dish to the incubator.
- Under a dissecting microscope, remove differentiated colonies from the dish to be passaged.
- Aspirate the spent medium from the dish with a Pasteur pipette, and rinse the dish once with Dulbecco’s PBS (DPBS) without Calcium and Magnesium.
- Add Collagenase Type IV (10 mg/mL) solution to the dish containing iPSCs. Adjust the volume of
Collagenase Type IV for various dish sizes (e.g., 35-mm dishes require 1 mL of Collagenase IV). - Incubate the dish(es) for 5–7 minutes in a 37°C, 5% CO2 incubator. Note that the incubation times may vary among different batches of collagenase; therefore, examination of the colonies is needed to determine the appropriate incubation time.
Note: As an alternative to Collagenase Type IV, you may use Dispase at a concentration of 2 mg/mL and incubate the dish(es) for 2–3 minutes in a 37° C, 5% CO2 incubator. - Stop the incubation when the edges of the colonies are starting to pull away from the plate (see Figure 3).
Figure 3. PSC colony pulling away from the Geltrex® matrix layer after treatment with Collagenase Type IV
- Aspirate the Collagenase Type IV Solution with a Pasteur pipette. Remove the collagenase carefully without disturbing the attached cell layer.
- Add MEF-CM to each dish. Use a 5-mL pipette to gently blow the cells off the surface of the dish while
pipetting up and down. Make sure to pipet gently to minimize the formation of bubbles. - After the iPSCs have been removed from the surface of the well, pool the contents of the wells into a 15-mL conical tube.
- Using a 5-mL pipette, add MEF-CM to the dish to wash and collect any residual cells. Pipet up the medium and cells, and then add the collected cells to the 15-mL tube.
- Pipet cells up and down gently a few times in the 15-mL tube to further break up cell colonies. Pipet
carefully to reduce foaming.
Note: Avoid making a single cell suspension. - Centrifuge at 200 × g for 5 minutes, and then aspirate the supernatant from the iPSC pellet.
- Resuspend the pellet with an appropriate amount of MEF-CM (refer to Table 2). This is dependent on the split ratio and the number of dishes used.
- Mix the cell suspension well with a 10-mL pipette. Be careful not to break up the colonies too much or cause bubbles in the media.
- Add appropriate volume of cell suspension to each dish. Return the dish to the incubator.
- Move the dish(es) in several quick, short, back-and-forth and side-to-side motions to disperse cells across the surface of the dishes.
- Incubate cells overnight to allow colonies to attach. Replace spent medium daily.
Note: While cells are attaching, be careful when opening and closing the incubator doors to avoid disturbing the even distribution of cells on the surface of the wells.
Mechanical Passaging Using the StemPro® EZPassage™ Disposable Cell Passaging Tool
- Replace the medium in the dish containing the cells with fresh MEF-CM.
- Under a laminar flow hood, open the package containing the EZPassage™ tool and remove the tool.
- Hold the culture vessel in one hand and pull (roll) the EZPassage™ tool across the entire dish in one direction. Apply gentle but firm pressure so that the entire roller blade touches the dish and maintains uniform pressure during the rolling action.
- Keep rolling the EZPassage™ tool parallel to the first pass until the entire dish has been covered.
- Rotate the culture dish 90°, and then repeat rolling the cell layer as described above.
- When you are finished, discard the EZPassage™ tool and do not reuse. Use a cell scraper to lift cell clusters off the plate, if necessary.
- Using a serological pipette, rinse the dish with medium so that the cut colonies are suspended in the medium.
- Transfer the medium containing the colonies to a 15-mL sterile tube.
- Seed the cell colonies on dishes plated with mitotically inactivated MEFs at an appropriate density.
- Place the plates into a 37°C, 5% CO2 incubator. Shake the plates gently to evenly spread out cells.
Figure 4. PSC Colony after being cut with the StemPro® EZPassage™ Disposable Cell Passaging Tool
- In the PSC Culture Medium, DMEM/F12 containing GlutaMAX™-I (Cat. no 10565-018) can be substituted with the following alternatives:
KnockOut™ DMEM/F-12 (Cat. no. 12660-012)
To prepare 100 mL of complete PSC Culture Medium using KnockOut™ DMEM/F-12, aseptically combine the components listed in the table below
Component | Stock Concentration | Final Concentration | Volume |
---|
KnockOut™ DMEM/F-12 (Cat. no. 12660-012) | — | 1X | 78 mL |
KnockOut™ SR (Cat. no. 10828-028) | — | 20% | 20 mL |
GlutaMAX™-I (Cat. no. 35050-061) | 200 mM | 2 mM | 1 mL |
MEM Non-Essential Amino Acids Solution (Cat. no. 11140-050) | 10 mM | 0.1 mM | 1 mL |
bFGF (Cat. no. PHG0024) | 10 μg/mL | 4 ng/mL | 40 μL |
β-mercaptoethanol (Cat. no. 21985-023) | 1000X | 1X | 100 μL |
KnockOut™ DMEM (Cat. no. 10829-018)
To prepare 100 mL of complete PSC Culture Medium using KnockOut™ DMEM, aseptically combine the components listed in the table below.
Component | Stock Concentration | Final Concentration | Volume |
---|
KnockOut™ DMEM (Cat. no. 10829-018) | — | 1X | 78 mL |
KnockOut™ SR (Cat. no. 10828-028) | — | 20% | 20 mL |
GlutaMAX™-I (Cat. no. 35050-061) | 200 mM | 2 mM | 1 mL |
MEM Non-Essential Amino Acids Solution (Cat. no. 11140-050) | 10 mM | 0.1 mM | 1 mL |
bFGF (Cat. no. PHG0024) | 10 μg/mL | 4 ng/mL | 40 μL |
β-mercaptoethanol (Cat. no. 21985-023) | 1000X | 1X | 100 μL |
Alternative bFGF pack sizes
Product Name | Cat. no. | Product Size |
---|
FGF-basic (AA 1-155) Recombinant Human | PHG0264 | 10 μg |
FGF-basic (AA 1-155) Recombinant Human | PHG0266 | 25 μg |
FGF-basic (AA 1-155) Recombinant Human | PHG0261 | 100 μg |
FGF-basic (AA 1-155) Recombinant Human | PHG0263 | 1 mg |
Dissociation Enzymes/Tools for Harvesting iPSC
Dissociation Enzyme /Tools | Application | Suggested concentration |
---|
StemPro® EZPassage™ tool (Cat. no. 23181-010) | Manual passaging | Sterile, disposable tool |
StemPro® Accutase® (Cat. no. A11105-01) | Monolayer of cells post passage, Dissociation into single cells | 1X ready to use (1–2 minutes incubation at 37°C) |
Dispase (Cat. no. 17105-041) | Colony-like morphology post passage | 2 mg/mL (2–3 minutes incubation at 37°C) |
TrypLE™ Express (Cat. no. 12604-021) | Dissociation to single cells | 1X ready to use |
TOP
For research use only. Not intended for any animal or human therapeutic or diagnostic use.