Thermo Scientific hydrides
To inorganic chemists, hydrides are compounds or ions made up of hydrogen bonded to an electropositive (or a less electronegative) element. Hydrides can be formed with almost all main-group elements, but inorganic hydrides are usually classified iinto four main groups: covalent, ionic, transition metal, and metallic hydrides. Ionic hydrides form between alkali and alkaline earth metals of group I and II metals like lithium, sodium, and calcium. Transition metal hydrides usually contain complexing ligands. Metallic hydrides or interstitial hydrides are formed by transition metals, lanthanides, or actinides.
Some of the popular reducing agents used for chemical transformations in research laboratories are metal borohydrides, metal aluminum hydrides, and diisobutylaluminium hydride (DIBAL). Superhydride, lithium triethylborohydride, is a strong and selective reducing agent. These hydrides reduce several functional groups including aldehydes, ketones, α,β-unsaturated carbonyl compounds, imines, nitro compounds, oximes, esters, lactones, amides, and acid anhydrides. Alkali hydrides are widely used in organic synthesis. Alkali earth metal hydrides like calcium hydride are effective desiccants for the removal of trace waters from aprotic solvents. Hydride complexes are widely used in a variety of homogeneous and heterogeneous catalysis, such as hydroformylation and hydrodesulfurization.
Thermo Fisher Scientific offers a range of hydrides in a variety of formats, including custom and bulk quantities.
Popular products
Catalog # Name Price (USD) Qty
041590.22 Calcium hydride, 92% min
132.00
047203.09 Zirconium hydride, 99%, Thermo Scientific Chemicals -
A13298.22 Tri-n-butyltin hydride, 97% -
L13266.ND Potassium hydride, 30% w/w in mineral oil
632.65
Online Exclusive
703.00
Save 70.35 (10%)
013442.14 Lithium aluminum hydride, 95% min
61.65
Online Exclusive
68.60
Save 6.95 (10%)
013442.36 Lithium aluminum hydride, 95% min
771.65
Special offer
Online exclusive
908.00
Save 136.35 (15%)
013442.59 Lithium aluminum hydride, 95% min
783.65
Online Exclusive
871.00
Save 87.35 (10%)
A13298.09 Tri-n-butyltin hydride, 97% -
A16242.30 Calcium hydride, coarse powder, ca 92%
193.65
Special offer
Online exclusive
228.00
Save 34.35 (15%)
012857.06 Titanium(II) hydride, 99% (metals basis)
37.90
041645.18 Calcium hydride, 98% (metals basis excluding Mg), Mg <1%
200.65
Special offer
Online exclusive
236.00
Save 35.35 (15%)
013442.51 Lithium aluminum hydride, 95% min
197.65
Online Exclusive
219.00
Save 21.35 (10%)
089643.14 Lithium aluminum hydride, pellets, 97%
66.65
Online Exclusive
73.60
Save 6.95 (9%)
089643.36 Lithium aluminum hydride, pellets, 97%
815.65
Online Exclusive
906.00
Save 90.35 (10%)
041596.22 Lithium hydride, 99.4% (metals basis)
352.00
A13298.30 Tri-n-butyltin hydride, 97% -
A16242.18 Calcium hydride, coarse powder, ca 92%
52.65
Special offer
Online exclusive
62.00
Save 9.35 (15%)
012857.22 Titanium(II) hydride, 99% (metals basis)
135.65
Online Exclusive
151.00
Save 15.35 (10%)
089183.06 Titanium(II) hydride, Ti 94.0% min.
28.10
019106.14 Calcium hydride, 90-95%
45.20
041645.30 Calcium hydride, 98% (metals basis excluding Mg), Mg <1%
879.65
Special offer
Online exclusive
1,035.00
Save 155.35 (15%)
A13298.18 Tri-n-butyltin hydride, 97% -
019106.22 Calcium hydride, 90-95%
108.65
Special offer
Online exclusive
128.00
Save 19.35 (15%)
A13298.14 Tri-n-butyltin hydride, 97% -
047203.18 Zirconium hydride, 99%, Thermo Scientific Chemicals -
A16242.0B Calcium hydride, coarse powder, ca 92%
619.65
Online Exclusive
688.00
Save 68.35 (10%)
030716.09 Lithium tri-tert-butoxyaluminum hydride, 94%
88.65
Special offer
Online exclusive
104.00
Save 15.35 (15%)
012857.D3 Titanium(II) hydride, 99% (metals basis)
1,935.65
Special offer
Online exclusive
2,277.00
Save 341.35 (15%)
012857.36 Titanium(II) hydride, 99% (metals basis)
556.65
Special offer
Online exclusive
655.00
Save 98.35 (15%)
013442.22 Lithium aluminum hydride, 95% min
130.65
Special offer
Online exclusive
154.00
Save 23.35 (15%)
L13266.NB Potassium hydride, 30% w/w in mineral oil
269.65
Special offer
Online exclusive
317.00
Save 47.35 (15%)
089183.30 Titanium(II) hydride, Ti 94.0% min.
459.00
013431.18 Sodium hydride, 57-63% oil dispersion
45.50
041596.14 Lithium hydride, 99.4% (metals basis)
107.65
Special offer
Online exclusive
126.00
Save 18.35 (15%)
013431.36 Sodium hydride, 57-63% oil dispersion
179.65
Special offer
Online exclusive
211.00
Save 31.35 (15%)
089643.22 Lithium aluminum hydride, pellets, 97%
188.65
Online Exclusive
209.00
Save 20.35 (10%)
089183.18 Titanium(II) hydride, Ti 94.0% min.
112.65
Online Exclusive
125.00
Save 12.35 (10%)
047203.30 Zirconium hydride, 99%, Thermo Scientific Chemicals -
041590.36 Calcium hydride, 92% min
386.00