basal cells or basal keratinocytes
Our Primary Human Keratinocyte Cell Cultures are optimized to synthesize keratinocyte cells, the predominant cell type in the epidermis. The keratinocytes found in the basal layer or stratum germinativum of the skin are sometimes referred to as basal cells or basal keratinocytes.
Primary Human Keratinocyte Cells adult (HEKa)
Primary Human Keratinocyte Cells neonatal (HEKn)
  • > 500,000 cells/vial cryopreserved at the end of the 1' culture
  • > 70% viability
  • > 25 population doublings HEKa (adult)
  • > 30 population doublings HEKn (neonatal)
  • Optimized for superior performance over the leading competitors
Phase contrast images of Human Epidermal Keratinocytes (HEKa) grown in culture
HEKa cells grown in EpiLife (M-EPI-500-CA) with the addition of Human Keratinocyte Growth Supplement (HKGS). First culture after thaw.
HEKa cells grown in EpiLife
Get more life out of you cells
Keratinocyte Cell Culture Graph
Gibco EpiLife serum-free cell culture medium
  • Defined, animal origin-free medium
  • Extends the in vitro lifespan of primary cells up to two times compared to other serum-free media
  • Ease of use with single-shot supplementation
Figure 1. Normal neonatal human keratinocytes (HEKn) (C-001-5C) were grown in EpiLife medium and in a keratinocyte medium from a leading competitor. Cultures grown in EpiLife medium demonstrated population doublings over an extended period compared to cells grown in the competitor's medium.
Keratinocyte product selection guide
If you'd like to culture your cells....Then choose:
  • in an animal-product free (APF) environment
  • for an extended lifespan
  • in a chemically-defined environment
  • for an extended lifespan
  • in a bovine pituitary extract (BPE)–containing environment
  • for an extended lifespan
  • in a bovine pituitary extract (BPE)–containing environment
  • for a standard lifespan
Ordering information
Catalog # Name Size Price (USD) Qty
C0055C Human Epidermal Keratinocytes, adult (HEKa) Each
724.00
Ordering information
Ordering information
Catalog # Name Size Price (USD) Qty
A1254201 Synth-a-Freeze™ Cryopreservation Medium Each
141.65
Online Exclusive
156.00
Save 14.35 (9%)
S001K Human Keratinocyte Growth Supplement Kit (HKGS Kit) Each
93.65
Online Exclusive
98.75
Save 5.10 (5%)
M154CF500 Medium 154CF, Kit Each
121.00
R01510 Gentamicin/Amphotericin Solution Each
117.00
R007100 Defined Trypsin Inhibitor Each
63.50
17005042 Keratinocyte SFM (1X) Each
152.65
Online Exclusive
167.00
Save 14.35 (9%)
M154CFPRF500 Medium 154CF/PRF, Kit Each
111.65
Online Exclusive
119.00
Save 7.35 (6%)
MEPI500CA EpiLife™ Medium, with 60 μM calcium Each
124.65
Online Exclusive
134.00
Save 9.35 (7%)
MEPICFPRF500 EpiLife™ CF/PRF Kit Each
138.00
R001100 Trypsin/EDTA Solution (TE) Each
41.30
M154500 Medium 154 Each
97.65
Online Exclusive
108.00
Save 10.35 (10%)
R002100 Trypsin Neutralizer Solution Each
38.65
Online Exclusive
41.28
Save 2.63 (6%)
10744019 Defined Keratinocyte SFM (1X) Each
144.65
Online Exclusive
161.00
Save 16.35 (10%)
A1371301 CTS™ Synth-a-Freeze™ Medium Each
249.00
MEPICF500 EpiLife™ CF Kit Each
139.00
S0175 Supplement S7 Each
125.00
S0125 EpiLife™ Defined Growth Supplement (EDGS) Each
89.75
S0015 Human Keratinocyte Growth Supplement (HKGS) Each
69.65
Online Exclusive
74.75
Save 5.10 (7%)
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions