Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Tissue culture tubes
      Choose from round and flat-bottom tubes as well as conical tubes that offer a variety of sizes and features, including recyclable racks and ease-of-use designs for enhanced performance.
      Centrifuge tubes  Storage tubes Request sample/quote
      Cell culture centrifuge tubes
      Approximately 24 conical bottomed centrifuge tube in a blue rack matching their blue caps
      Available in a wide variety of sizes, colors, and formats, Nunc centrifuge tubes are designed to meet a range of needs. Navigate centrifuge offerings below by size.
        Tubes brochure
        Read now how our protein expression systems perform bioreactor tubes
      EZFlip conical centrifuge tubes
      A blue-gloved hand holding an EZFlip tube demonstrating its ability to be opened one handed
      Nunc 15 mL EZFlip conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Integrated leakproof cap
      • Writing area for sample identification
      Nunc 50 mL EZFlip conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Integrated leakproof cap
      • Writing area for sample identification
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      362694Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalNoNo50500Order
      362694NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes50500Order
      362695Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalYesYes50500Order
      362695NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalYesNo50500Order
      362696NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoNo25500Order
      362697NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes25500Order
      Return to top of tab
      Bioreactor conical centrifuge tubes
      Three bioreactor tubes, one laying on its side
      Nunc 50 mL bioreactor tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      • Four different sizes of ventilation (dial type) on the cap
      • 2 µm PTFE filter membrane
        Read now how our protein expression systems perform bioreactor tubes
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      332260Nunc 50 mL Bioreactor Tube UntreatedConicalNoYes390Order
      Return to top of tab
      15 mL-500 mL conical centrifuge tubes
      5 conical bottomed centrifuge tube in a blue rack matching their blue caps
      Nunc 15 mL conical centrifuge tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      A white gloved hand on white background holding a conical bottomed 50mL tube
      Nunc 50 mL conical centrifuge tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      Media being pipetted into a single wide-mouthed 250mL conical centrifuge tube
      Nunc 250 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      • Wide-mouth design
      A single 250mL conical centrifuge tube with light blue cap
      250 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      A single 500mL conical centrifuge tube with light blue cap
      500 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • DNase-/RNase-free
      • BSE-/TSE-free
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      339650Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes50500Order
      339651Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25500Order
      339652Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes25500Order
      339653Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25300Order
      376814250 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
      332265250 mL Conical Centrifuge TubesUntreatedConicalNoYes648Order
      332270500 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
      Return to top of tab
      EZFlip conical centrifuge tubes
      A blue-gloved hand holding an EZFlip tube demonstrating its ability to be opened one handed
      Nunc 15 mL EZFlip conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Integrated leakproof cap
      • Writing area for sample identification
      Nunc 50 mL EZFlip conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Integrated leakproof cap
      • Writing area for sample identification
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      362694Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalNoNo50500Order
      362694NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes50500Order
      362695Nunc 15 mL EZFlip Conical Centrifuge TubesUntreatedConicalYesYes50500Order
      362695NSNunc 15 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalYesNo50500Order
      362696NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoNo25500Order
      362697NSNunc 50 mL EZFlip Conical Centrifuge Tubes (Non-sterile)UntreatedConicalNoYes25500Order
      Return to top of tab
      Bioreactor conical centrifuge tubes
      Three bioreactor tubes, one laying on its side
      Nunc 50 mL bioreactor tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      • Four different sizes of ventilation (dial type) on the cap
      • 2 µm PTFE filter membrane
        Read now how our protein expression systems perform bioreactor tubes
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      332260Nunc 50 mL Bioreactor Tube UntreatedConicalNoYes390Order
      Return to top of tab
      15 mL-500 mL conical centrifuge tubes
      5 conical bottomed centrifuge tube in a blue rack matching their blue caps
      Nunc 15 mL conical centrifuge tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      A white gloved hand on white background holding a conical bottomed 50mL tube
      Nunc 50 mL conical centrifuge tubes feature:
      • Virgin-polypropylene-resin composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      Media being pipetted into a single wide-mouthed 250mL conical centrifuge tube
      Nunc 250 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      • Wide-mouth design
      A single 250mL conical centrifuge tube with light blue cap
      250 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • Leakproof guarantee
      A single 500mL conical centrifuge tube with light blue cap
      500 mL conical centrifuge tubes feature:
      • Clarified polypropylene composition
      • Nonpyrogenic and non-cytotoxic guarantee
      • DNase-/RNase-free
      • BSE-/TSE-free
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      339650Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes50500Order
      339651Nunc 15 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25500Order
      339652Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalNoYes25500Order
      339653Nunc 50 mL Conical Sterile Polypropylene Centrifuge TubesUntreatedConicalYesYes25300Order
      376814250 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
      332265250 mL Conical Centrifuge TubesUntreatedConicalNoYes648Order
      332270500 mL Conical Centrifuge TubeUntreatedConicalNoYesNA40Order
      Return to top of tab
      Nunclon Delta culture tubes for fastidious cells
      Nunclon Delta tube surface for adherent cells
      The Nunclon Delta surface treatment for adherent cells, makes standard polystyrene surfaces more hydrophilic, facilitating in maximum adhesion for a range of cell types.
      With this culture tube surface treatment, you can perform adherent cell cultures in biotechnology, pharmaceutical, and research labs as well as with vaccine development and diagnostic kits.
      Explore cell performance with Nunclon Delta surfaces
      Cat. No.DescriptionSurfaceTube shapeCapSterilePack sizeCase 
      156758
      Nunc Cell Culture Tubes (3 mL)
      Nunclon Delta
      FlatScrew capYes75450Order
      145470
      Nunc Cell Culture Tubes (7 mL)
      Nunclon Delta
      RoundPush-onYes100600Order
      146183
      Nunc Cell Culture Tubes (7 mL)
      Nunclon Delta
      RoundScrew capYes100600Order
      Cell culture storage tubes and racks
      Nunc storage tubes for tissue culture
      Available in a wide variety of sizes, colors, and formats Nunc Storage Tubes provide storage tubes for all your needs.  Matrix 2D tube technologies securely store samples at low temperatures, including vapor phase liquid nitrogen. A variety of racks for your choice of storage tube are arranged below by size.
      Order Matrix ScrewTop tube caps, cap trays, and cap inserts
      500 µL storage tubes
      A storage rack holding a variety of 500 µL tubes with various colored caps
      Matrix 500 µL V-bottom storage tubes feature:
      • Virgin-polypropylene-resin composition
      • ScrewTop caps with soft silicone gaskets
      • Stringent 2D barcode quality control
      Order matrix storage tube racks
      Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
      3743
      Matrix 500 μL ScrewTop Tubes
      Colorless
      V-bottomNoYesNA480Order
      3744
      Matrix 500 μL ScrewTop Tubes
      Colorless
      V-bottomYesYes96/rack480Order
      3743RED
      Matrix 500 μL ScrewTop Tubes
      Red
      V-bottomNoYesNA480Order
      3744RED
      Matrix 500 μL ScrewTop Tubes
      Red
      V-bottomYesYes96/rack480Order
      3743YEL
      Matrix 500 μL ScrewTop Tubes
      Yellow
      V-bottomNoYesNA480Order
      3744YEL
      Matrix 500 μL ScrewTop Tubes
      Yellow
      V-bottomYesYes96/rack480Order
      3743GRE
      Matrix 500μL ScrewTop Tubes
      Green
      V-bottomNoYesNA480Order
      3743BLU
      Matrix 500 μL ScrewTop Tubes
      Blue
      V-bottomNoYesNA480Order
      3743PUR
      Matrix 500 μL ScrewTop Tubes
      Purple
      V-bottomNoYesNA480Order
      3743WHI
      Matrix 500 μL ScrewTop Tubes
      White
      V-bottomNoYesNA480Order
      0.75 mL storage tubes
      A storage rack filled with 0.75 mL tubes
      Matrix 0.75 mL blank and alphanumeric storage tubes feature:
      • Virgin-polypropylene-resin composition
      • Alphanumeric codes on tube bottoms
      • A variety of cap options
      Order matrix storage tube racks
      Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
      4170
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoNoNA1000Order
      4272
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      YesYes10/rack960Order
      4273
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      YesNo10/rack960Order
      1 mL storage tubes
      A storage rack filled with 1 mL tubes with various cap colors
      Matrix 1 mL V-bottom storage tubes feature:
      • Virgin-polypropylene-resin composition
      • ScrewTop caps with soft silicone gaskets
      • Stringent 2D barcode quality control
      Order matrix storage tube racks
      Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
      3741
      Matrix 1.0 mL ScrewTop Tubes
      Colorless
      V-bottomYesYes96/rack480Order
      3740RED
      Matrix 1.0 mL ScrewTop Tubes
      Red
      V-bottomNoYesNA480Order
      3741RED
      Matrix 1.0 mL ScrewTop Tubes
      Red
      V-bottomYesYes96/rack480Order
      3740YEL
      Matrix 1.0 mL ScrewTop Tubes
      Yellow
      V-bottomNoYesNA480Order
      3741YEL
      Matrix 1.0 mL ScrewTop Tubes
      Yellow
      V-bottomYesYes96/rack480Order
      3740GRE
      Matrix 1.0 mL ScrewTop Tubes
      Green
      V-bottomNoYesNA480Order
      3740BLU
      Matrix 1.0 mL ScrewTop Tubes
      Blue
      V-bottomNoYesNA480Order
      3741BLU
      Matrix 1.0 mL ScrewTop Tubes
      Blue
      V-bottomYesYes96/rack480Order
      3740PUR
      Matrix 1.0 mL ScrewTop Tubes
      Purple
      V-bottomNoYesNA480Order
      3740WHI
      Matrix 1.0 mL ScrewTop Tubes
      White
      V-bottomNoYesNA480Order
      1.4 mL storage tubes
      A storage rack filled with 1.4 mL tubes
      Matrix 1.4 mL blank and alphanumeric storage tubes feature:
      • Virgin-polypropylene-resin composition
      • Alphanumeric codes on tube bottoms
      • A variety of cap options
      Order matrix storage tube racks
      Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
      4253
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      NoNo10/carrier960Order
      4140
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoNoNA1000Order
      4147
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      YesNo10/rack960Order
      4250
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoYes10/carrier960Order
      Return to top of tab
      12 mL storage culture tubes
      A single unfilled 12 mL storage tube on white background
      Matrix 12 mL storage tubes feature:
      • Laser-etched 2D barcode on tube bottom
      • Tested against leaks
      • Built-in error correction
      Order matrix storage tube racks
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      3778
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundNoYes4/2496Order
      3776
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNA4Order
      3777TS
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNAEachOrder
      3770
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundNoNoNA100Order
      3774
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNAEachOrder
      3775
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesYes4/2496Order
      500 µL storage tubes
      A storage rack holding a variety of 500 µL tubes with various colored caps
      Matrix 500 µL V-bottom storage tubes feature:
      • Virgin-polypropylene-resin composition
      • ScrewTop caps with soft silicone gaskets
      • Stringent 2D barcode quality control
      Order matrix storage tube racks
      Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
      3743
      Matrix 500 μL ScrewTop Tubes
      Colorless
      V-bottomNoYesNA480Order
      3744
      Matrix 500 μL ScrewTop Tubes
      Colorless
      V-bottomYesYes96/rack480Order
      3743RED
      Matrix 500 μL ScrewTop Tubes
      Red
      V-bottomNoYesNA480Order
      3744RED
      Matrix 500 μL ScrewTop Tubes
      Red
      V-bottomYesYes96/rack480Order
      3743YEL
      Matrix 500 μL ScrewTop Tubes
      Yellow
      V-bottomNoYesNA480Order
      3744YEL
      Matrix 500 μL ScrewTop Tubes
      Yellow
      V-bottomYesYes96/rack480Order
      3743GRE
      Matrix 500μL ScrewTop Tubes
      Green
      V-bottomNoYesNA480Order
      3743BLU
      Matrix 500 μL ScrewTop Tubes
      Blue
      V-bottomNoYesNA480Order
      3743PUR
      Matrix 500 μL ScrewTop Tubes
      Purple
      V-bottomNoYesNA480Order
      3743WHI
      Matrix 500 μL ScrewTop Tubes
      White
      V-bottomNoYesNA480Order
      0.75 mL storage tubes
      A storage rack filled with 0.75 mL tubes
      Matrix 0.75 mL blank and alphanumeric storage tubes feature:
      • Virgin-polypropylene-resin composition
      • Alphanumeric codes on tube bottoms
      • A variety of cap options
      Order matrix storage tube racks
      Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
      4170
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoNoNA1000Order
      4272
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      YesYes10/rack960Order
      4273
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      YesNo10/rack960Order
      1 mL storage tubes
      A storage rack filled with 1 mL tubes with various cap colors
      Matrix 1 mL V-bottom storage tubes feature:
      • Virgin-polypropylene-resin composition
      • ScrewTop caps with soft silicone gaskets
      • Stringent 2D barcode quality control
      Order matrix storage tube racks
      Cat. No.DescriptionColorTube shapeWith rackSterilePack sizeCase 
      3741
      Matrix 1.0 mL ScrewTop Tubes
      Colorless
      V-bottomYesYes96/rack480Order
      3740RED
      Matrix 1.0 mL ScrewTop Tubes
      Red
      V-bottomNoYesNA480Order
      3741RED
      Matrix 1.0 mL ScrewTop Tubes
      Red
      V-bottomYesYes96/rack480Order
      3740YEL
      Matrix 1.0 mL ScrewTop Tubes
      Yellow
      V-bottomNoYesNA480Order
      3741YEL
      Matrix 1.0 mL ScrewTop Tubes
      Yellow
      V-bottomYesYes96/rack480Order
      3740GRE
      Matrix 1.0 mL ScrewTop Tubes
      Green
      V-bottomNoYesNA480Order
      3740BLU
      Matrix 1.0 mL ScrewTop Tubes
      Blue
      V-bottomNoYesNA480Order
      3741BLU
      Matrix 1.0 mL ScrewTop Tubes
      Blue
      V-bottomYesYes96/rack480Order
      3740PUR
      Matrix 1.0 mL ScrewTop Tubes
      Purple
      V-bottomNoYesNA480Order
      3740WHI
      Matrix 1.0 mL ScrewTop Tubes
      White
      V-bottomNoYesNA480Order
      1.4 mL storage tubes
      A storage rack filled with 1.4 mL tubes
      Matrix 1.4 mL blank and alphanumeric storage tubes feature:
      • Virgin-polypropylene-resin composition
      • Alphanumeric codes on tube bottoms
      • A variety of cap options
      Order matrix storage tube racks
      Cat. No.DescriptionBarcodeWith latch rackSterilePack sizeCase 
      4253
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      NoNo10/carrier960Order
      4140
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoNoNA1000Order
      4147
      Matrix Blank and Alphanumeric Storage Tubes
      Alphanumeric
      YesNo10/rack960Order
      4250
      Matrix Blank and Alphanumeric Storage Tubes
      Blank
      NoYes10/carrier960Order
      Return to top of tab
      12 mL storage culture tubes
      A single unfilled 12 mL storage tube on white background
      Matrix 12 mL storage tubes feature:
      • Laser-etched 2D barcode on tube bottom
      • Tested against leaks
      • Built-in error correction
      Order matrix storage tube racks
      Cat. No.DescriptionSurfaceTube shapeWith rackSterilePack sizeCase 
      3778
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundNoYes4/2496Order
      3776
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNA4Order
      3777TS
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNAEachOrder
      3770
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundNoNoNA100Order
      3774
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesNoNAEachOrder
      3775
      Matrix 2D Barcoded 12 mL Screw-top Storage Tubes
      Untreated
      RoundYesYes4/2496Order