EDTA (0.5 M), pH 8.0
EDTA (0.5 M), pH 8.0
Thermo Scientific™

EDTA (0.5 M), pH 8.0

Thermo Scientific 0.5 M EDTA, pH 8.0 is an aqueous solution of 0.22 μm membrane-filtered ethylendiaminetetraacetic acid (EDTA), sodium salt.Read more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
R1021Promo Image5 x 1 mL
Catalog number R1021
Price (USD)
24.65
Online Exclusive
30.26
Save 5.61 (19%)
Each
In stock
Add to cart
Quantity:
5 x 1 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
24.65
Online Exclusive
30.26
Save 5.61 (19%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific 0.5 M EDTA, pH 8.0 is an aqueous solution of 0.22 μm membrane-filtered ethylendiaminetetraacetic acid (EDTA), sodium salt. It's ideal for biochemistry or molecular biology applications that require a chelator for divalent cations. By forming complexes with metal ions, EDTA inhibits metal-dependent enzymatic reactions. EDTA complexes metal ions at a 1:1 molar ratio.

Nucleic acids are normally stored in TE buffer that contains 1 mM EDTA, which prevents degradation of nucleic acids by metal dependent nucleases.

Applications

• Termination of metal dependent enzymatic reactions
• Protection of DNA against metal dependent nucleases
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialEDTA
Product TypeEDTA
Quantity5 x 1 mL
FormLiquid
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3224598Certificate of AnalysisApr 24, 2025R1021
3224601Certificate of AnalysisApr 23, 2025R1021
3207512Certificate of AnalysisApr 09, 2025R1021
3191910Certificate of AnalysisMar 18, 2025R1021
3190909Certificate of AnalysisMar 17, 2025R1021
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link