Improve your cell culture with Gibco media supplements 
Media supplements are often a vital component of successful cell culture experiments. There are many advantages of using cell culture supplements including customizing the growth conditions of your cells, improving cell viability and growth, and keeping cells healthier longer.
Gibco cell culture media supplements help ensure the reliability and consistency of your cell culture and research results.
Variety of bottles containing antibiotics products
Antibiotics 
Proper supplementation of cell culture media with antibiotics can protect your cells from contamination. In addition, certain selection antibiotics are used for stable transfection and the development of cell lines.
Explore cell culture antibiotics
Bottles containing L-glutamine and GlutaMAX supplements
L-glutamine & GlutaMAX supplement
L-glutamine is an essential supplement providing energy to rapidly dividing cells. Gibco GlutaMAX offers enhanced stability and supports improved cell health.
Explore L-glutamine and GlutaMAX supplement
product photo
Insulin-Transferrin-Selenium
Routinely used with basal media to support a wide variety of cell types in reduced sera or serum-free cell culture conditions. Animal origin and animal origin-free ITS formulations are available.
Explore ITS supplements
Product photo of Gibco HEPES
Cell culture buffers
It is important to provide a buffer to cells to protect against changes in pH. This essential buffering is achieved by including HEPES or sodium bicarbonate based buffer in your cell culture media.
Explore cell culture buffers
Tubes containing bovine serum albumin (BSA)
Bovine serum albumin (BSA)
BSA provides cells in culture with enhanced nutrition, reducing the need for supplementation with serum. BSA is also useful in other laboratory workflows, including as a blocking reagent for immunoassays.
Explore bovine serum albumin (BSA)
Product photo of Gibco HEPES
Growth factors & cytokines
Growth factors and cytokines are important for the proliferation and development of cells in culture. Gibco PeproTech recombinant growth factors and cytokines offer quality and consistency to support robust cell culture and disease models.
Explore growth factors and cytokines
Additional media supplements
Insulin-Transferrin-Selenium
Gibco Insulin-Transferrin-Selenium (ITS) is a basal medium supplement that helps increase cell proliferation and viability while reducing the amount of fetal bovine serum used to culture cells. ITS supplements provide cells with some of the essential components naturally found in FBS.
We offer an array of ITS formulations to support the unique needs of the diverse cell types researchers work with every day. The original ITS formulation (ITS-G) can be used to support a variety of cell types while more specific formulations offer distinct advantages for specific cell types. Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) supports cell types that require ethanolamine supplementation, such as bronchial epithelial cells. Meanwhile, Insulin-Transferrin-Selenium-Sodium Pyruvate (ITS-A) provides an additional source of the important metabolite pyruvate.
Insulin-Transferrin-Selenium Select supplements for AOF applications
To support cell culture applications requiring a reduction or removal of animal origin materials, we offer Gibco ITS-G Select and ITS-X Select supplements. The components of these 100X supplements are animal origin–free and can be included in complete culture media formulations to support the growth and culture of many cell types.
Insulin-Transferrin-Selenium Selection Table
 ITS-GITS-XITS-AITS-G SelectITS-X Select
Cat. No.414000455150005651300044A4000046501
A4000046502
A4000046401
A4000046402
Animal origin
Indicates source of insulin and transferrin, whether derived from animal or non-animal sources.
Animal originAnimal originAnimal originanimal origin-free formulationanimal origin-free formulation
Insulin
Promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids.
Transferrin
An iron carrier and may also help to reduce toxic levels of oxygen radicals and peroxide.
Selenium
As sodium selenite is a co-factor for glutathione peroxidase and other proteins that is used as an anti-oxidant in media.
Ethanolamine
A precursor of phospho-glycerides which are essential to the structure of the plasma membrane and cellular organelles.
Sodium pyruvate
A precursor to amino acids, fatty acids, and cholesterol; it is an important intermediate in many biosynthetic pathways.
Cell culture buffers
Bottle containing Sodium Bicarbonate 7.5% solution
Sodium bicarbonate used in cell culture
Sodium bicarbonate (NaHCO3) is a common buffer used in cell culture for maintaining the pH of the medium in the presence of 4–10% carbon dioxide. In addition, sodium bicarbonate provides some nutritional benefits and is rarely toxic to cells.
The final concentration of sodium bicarbonate in media depends on both the formulation of the media and carbon dioxide concentration of the incubator. Different CO2 concentrations are recommended depending on the concentration of sodium bicarbonate used. If the media formulation contains:
  • NaHCO3 (g/L) <1.5, it needs CO2 at 4%
  • NaHCO3 (g/L) 1.5–2.2, it needs CO2 at 5%
  • NaHCO3 (g/L) 2.2–3.4, it needs CO2 at 7%
  • NaHCO3 (g/L) >3.5, it needs CO2 at 10%
Catalog # Name Size Price (EUR) Qty
25080094 Sodium Bicarbonate 7.5% solution Each
17,56
Bottle containing Gibco HEPES buffer
HEPES buffer used in cell culture
Although bicarbonate is the most commonly used cell culture buffer due to its nutritional benefits, the buffering capacity of bicarbonate is reduced at physiological pH compared to HEPES buffer. HEPES has no nutritional benefits to cells but can be added to buffer cell culture media at pH 7.2 through 7.6. Therefore, the addition of HEPES can allow for extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator.
Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent and is categorized as a "Good" buffer which derives from a set of buffers described by Dr. Norman Good and his colleagues in 1966 (Good, et al. Biochemistry 1966). 
"Good buffers" such as HEPES are attributed with the following characteristics:
  • pKa values between 6.0 and 8.0
  • High solubility
  • Membrane impermeability
  • Limited effect on biochemical reactions
  • Very low visible and ultraviolet light absorbance
  • Chemically and enzymatically stable
  • Easy to prepare
The level of HEPES in cell culture media may vary from 10 mM to 25 mM. In Gibco DMEM, HEPES is present at a concentration of 25 mM. However, in Gibco DMEM/F-12, it is found at a concentration of 15 mM. The most commonly used concentration is 25 mM. 
Catalog # Name Size Price (EUR) Qty
15630106 HEPES (1 M), 20 mL Each
58,00
15630080 HEPES (1 M), 100 mL Each
138,00
11344041 HEPES (Ultra Pure) Each
972,00
Other cell culture supplements 
2-mercaptoethanol
The widely used chemical compound 2-mercaptoethanol has the formula HOCH2CH2SH. It is a hybrid of ethylene glycol and is used to reduce disulfide bonds. By scavenging hydroxyl radicals (amongst others), it can act as a biological antioxidant. The hydroxyl group confers solubility in water and lowers volatility.
Catalog # Name Size Price (EUR) Qty
21985023 2-Mercaptoethanol Each
50,65
Online Exclusive
54,00
Save 3,35 (6%)
31350010 2-Mercaptoethanol (50 mM) Each
14,18
Product photo of Gibco MEM NEAA
Amino acid solutions 
To prolong the viability of cells in culture and stimulate growth, supplements of amino acids can be added to media. In addition, adding an amino acid supplement provides nutrients and reduces the biosynthetic burden on cells in vitro. Prepared in distilled water, the non-essential amino acids in this solution are 100X the concentration in a MEM-alpha medium.
HAT supplement 
HAT supplement is a liquid mixture of sodium hypoxanthine, aminopterin, and thymidine. After dilution, HAT is suitable for use as a post-fusion selective medium to eliminate unfused or self-fused HGPRT-myeloma cells. Hypoxanthine and thymidine provide preformed purines and pyrimidines for DNA synthesis by hybridomas via the salvage pathway that utilizes HGPRT—contributed by the fused spleen cell. The folic acid antagonist, aminopterin, inhibits the de novo nucleoside biosynthesis pathway.
Catalog # Name Size Price (EUR) Qty
21060017 HAT Supplement (50X) Each
36,97
Product photo of Gibco HT Supplement
HT supplement 
HT supplement liquid is a mixture of sodium hypoxanthine and thymidine. After dilution, HT supplement is suitable for use as a rescue medium. HT supplement provides preformed purines and pyrimidines to overcome the effects of residual intracellular aminopterin. Following reestablishment of the de novo biosynthesis pathway, HT supplementation can be discontinued.
Catalog # Name Size Price (EUR) Qty
11067030 HT Supplement (100X) Each
53,65
Online Exclusive
55,00
Save 1,35 (2%)
MEM vitamin solution 
Adding MEM vitamin solution to media can enhance cell growth and viability in culture and reduce biosynthetic burden on cells in vitro. Prepared in 0.85% saline, the vitamins in this solution are 100X the concentration in a standard MEM medium. 
Catalog # Name Size Price (EUR) Qty
11120052 MEM Vitamin Solution (100X) Each
29,28
11120037 MEM Vitamin Solution (100X) Each
21,26
Product photo of Gibco Sodium Pyruvate
Sodium pyruvate 
Sodium pyruvate is often supplemented into cell culture media as an additional source of carbon. Gibco sodium pyruvate for cell culture is supplied as a 100 mM (100X) solution with a final concentration of 11,004 mg/L.
Catalog # Name Size Price (EUR) Qty
11360088 Sodium Pyruvate (100 mM), 20 x 100 mL Each
245,00
11360039 Sodium Pyruvate (100 mM), 100 mL Each
14,11
11360070 Sodium Pyruvate (100 mM) Each
17,65
Online Exclusive
18,51
Phytohemagglutinin
Phytohemagglutinin is a solution produced from a crude extract of the red kidney bean Phaseolus vulgaris. When applied at certain concentrations, this supplement can induce blastogenesis in various mammalian mononuclear cells in vitro.
Catalog # Name Size Price (EUR) Qty
10576015 Phytohemagglutinin, M form (PHA-M) Each
67,25
Try the Gibco MediaSelect Tool
Easily find a media solution suited to your specific application. Use this interactive tool to select classical basal, protein expression, or viral vector media.
Your selections will allow you to choose from a catalog menu of media – or customize your own formulation to fit your experimental setup and budget. Where available, you can compare media formulations side-by-side.
Find your media with the Gibco MediaSelect Tool
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Gibco Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.