Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Membranes and Filter Papers for Western Blotting |
PVDF (polyvinylidene difluoride) and nitrocellulose membranes are used for transferring proteins following gel electrophoresis for subsequent immunodetection. Compare the various membranes available for western blotting and read about various considerations when choosing the right membrane for your experiment.
Protein transfer is easy and convenient with our pre-cut, pre-assembled western blot membrane/filter paper sandwiches designed to fit your gels. Alternatively, membrane rolls, sheets, and filters papers are available to enable individual sizing and assembly.
Format | Great for | Ease of use | Nitrocellulose | PVDF | Low fluorescence PVDF |
---|---|---|---|---|---|
Roll | Flexible membrane dimensions | + | Nitrocellulose Membranes, 0.45 µm, 1 Roll (Cat. No. 88018) | PVDF Transfer Membranes, 0.2 µm, 1 Roll (Cat. No. 88520) PVDF Transfer Membranes, 0.45 µm, 1 Roll (Cat. No. 88518) | Low Fluorescence PVDF Transfer Membranes, 0.3 µm, 1 Roll (Cat. No. 678000) |
Pre-cut sheets | Consistency, reduced risk of damage due to handling, reduced waste | ++ | Mini: Nitrocellulose Membranes, 0.2 µm, 8 x 8 cm (Cat. No. 88024) Mini: Nitrocellulose Membranes, 0.45 µm, 8 x 8 cm (Cat. No. 88025) Midi: Nitrocellulose Membranes, 0.2 µm, 8 x 12 cm (Cat. No. 77012) Midi: Nitrocellulose Membranes, 0.45 µm, 8 x 12 cm (Cat. No. 77010) | PVDF Transfer Membranes, 0.45 µm, 10 x 10 cm (Cat. No. 88585) | Mini: Low Fluorescence PVDF Membranes, 0.3µm, 8.3 x 7.3 cm (Cat. No. 678003) Midi: Low Fluorescence PVDF Membranes, 0.3µm, 13.5 x 8 cm (Cat. No. 678004) |
Membrane/filter paper sandwich | Ready-to-use for wet transfer, no assembly required | +++ | Mini: Nitrocellulose/Filter Paper Sandwich, 0.2 µm, 8.3 x 7.3 cm (Cat. No. LC2000) Mini: Nitrocellulose/Filter Paper Sandwich, 0.45 µm, 8.3 x 7.3 cm (Cat. No. LC2001) Midi: Nitrocellulose/Filter Paper Sandwich, 0.2 µm, 8.5 x 13.5 cm (Cat. No. LC2009) Midi: Nitrocellulose/Filter Paper Sandwich, 0.45 µm, 8.6 x 13.5 cm (Cat. No. LC2006) | Mini: PVDF/Filter Paper Sandwich, 0.2 µm, 8.3 x 7.3 cm (Cat. No. LC2002) Mini: PVDF/Filter Paper Sandwich, 0.45 µm, 8.3 x 7.3 cm (Cat. No. LC2005) Midi: PVDF/Filter Paper Sandwich, 0.45 µm, 8.5 x 13.5 cm (Cat. No. LC2007) | Mini: Low Fluorescence PVDF/Filter Paper Sandwich, 0.3µm, 8.3 x 7.3 cm (Cat. No. 678001) Midi: Low Fluorescence PVDF/Filter Paper Sandwich, 0.3µm, 13.5 x 8 cm (Cat. No. 678002) |
For rapid dry and semi-dry transfer systems, like the iBlot 3 Western Blot Transfer System and Power Blotter Semi-Dry Transfer System, specialized transfer stacks are available.
Transfer System | Best for | Ease of use | Nitrocellulose | PVDF | Low fluorescence PVDF |
---|---|---|---|---|---|
iBlot 3 | Fastest setup, clean-up, and transfer time (3–8 minutes) | +++++ | Mini: iBlot 3 Transfer Stacks, Nitrocellulose, 0.2 µm, 8 x 8 cm (Cat. No. IB33002) Midi: iBlot 3 Transfer Stacks, Nitrocellulose, 0.2 µm, 13.5 x 8 cm (Cat. No. IB33001) | Mini: iBlot 3 Transfer Stacks, PVDF, 0.2 µm, 8 x 8 cm (Cat. No. IB34002) Midi: iBlot 3 Transfer Stacks, PVDF, 0.2 µm, 13.5 x 8 cm (Cat. No. IB34001) | Mini: iBlot 3 Transfer Stacks, Low Fluorescence PVDF, 0.3 µm, 8 x 8 cm (Cat. No. IB34004) Midi: iBlot 3 Transfer Stacks, Low Fluorescence PVDF, 0.3 µm, 13.5 x 8 cm (Cat. No. IB34003) |
Power Blotter | Rapid transfer (5–10 minutes), flexible membrane and filter paper options | ++++ | Mini: Power Blotter Select Transfer Stacks, Nitrocellulose, 0.2 µm, 8 x 8 cm (Cat. No. PB3210) Mini: Power Blotter Pre-cut Membranes and Filters, Nitrocellulose, 0.2 µm, 7 x 8.4 cm (Cat. No. PB7220) Midi: Power Blotter Select Transfer Stacks, Nitrocellulose, 0.2 µm, 13 x 8.3 cm (Cat. No. PB3340) Midi: Power Blotter Pre-cut Membranes and Filters, Nitrocellulose, 0.2 µm, 13 x 8.5 cm (Cat. No. PB7320) | Mini: Power Blotter Select Transfer Stacks, PVDF, 0.2 µm, 8 x 8 cm (Cat. No. PB5240) Mini: Power Blotter Pre-cut Membranes and Filters, PVDF, 0.45 µm, 7 x 8.4 cm (Cat. No. PB9220) Midi: Power Blotter Select Transfer Stacks, PVDF, 0.2 µm, 13 x 8.3 cm (Cat. No. PB5310) Midi: Power Blotter Pre-cut Membranes and Filters, PVDF, 0.45 µm, 13.5 x 8.5 cm (Cat. No. PB9320) | The low fluorescence PVDF membranes listed in the above table are compatible with Power Blotter. |
High-quality western blotting results can often be achieved using either nitrocellulose or PVDF membranes. Both types of membranes offer efficient protein transfer and strong protein binding, enabling consistent results and sensitive detection for chemiluminescent western blotting. However, this can be highly protein dependent. Certain proteins bind more tightly to one type of membrane over the other. Broad assertions, such as better performance based on high or low molecular weight proteins, can be misleading or inaccurate. A protein’s properties (i.e., charge, hydrophobicity, etc.) affect its ability to bind to membrane surfaces; so, finding the optimal membrane may require experimenting with your specific protein on various membranes.
The detection method used can also impact the target signal and membrane background. In fluorescent western blotting, or when using fluorescent total protein normalization methods such as Invitrogen No-Stain Protein Labeling Reagent, exposure times are often limited due to membrane auto-fluorescence and background noise, which negatively impact the achievable limit of detection. Optimized low fluorescence PVDF membranes exhibit low auto-fluorescence across fluorescent channels enabling low background and high signal to noise, making them excellent for fluorescent-based western blotting applications.
Lastly, there are handling considerations that may influence the decision to use nitrocellulose or PVDF. PVDF membranes are generally considered more durable than nitrocellulose. However, PVDF requires an activation step to enable good protein binding. This step is usually performed through a 3–5-minute incubation in 100% methanol or ethanol. In contrast, nitrocellulose membranes do not require this pre-activation step but can be more brittle than PVDF and require more careful handing to avoid tearing.
Property | Nitrocellulose membrane | PVDF membrane | Low fluorescence PVDF membrane |
---|---|---|---|
Chemiluminescent detection | +++ | +++ | +++ |
Fluorescent detection | ++ | + | +++ |
Total protein normalization | ++ | + | +++ |
Requires activation? | No | Yes | Yes |
Binding mechanism | Nitrogen dipole, H-bond, ionic, and hydrophobic interactions | Hydrophobic interactions | Hydrophobic interactions |
Durability | Less durable | More durable | More durable |
Nitrocellulose membranes are a popular matrix used in protein blotting because of their high protein-binding affinity, compatibility with a variety of detection methods, and the ability to immobilize proteins, glycoproteins, and nucleic acids. These membranes are composed of nitrocellulose, a cellulose derivative that helps provide a robust and reliable medium for protein immobilization. Nitrocellulose membranes are compatible with colorimetric, chemiluminescent, and most fluorescent detection methods. Their ease of use and compatibility with various staining and detection protocols make these membranes a great choice for western blotting.
PVDF (polyvinylidene difluoride) membranes are hydrophobic, microporous membranes that are an ideal choice for western blotting applications, as well as for amino acid analysis and protein sequencing of small amounts of proteins (as little as 10 pmoles). In western blotting, PVDF membranes bind proteins through hydrophobic and dipole interactions.
PVDF membranes require an activation step prior to submersion in transfer buffer. This activation step is necessary due to the highly hydrophobic characteristics of PVDF. During the activation step, methanol or ethanol serves as a liaison between the non-polar membrane and the polar buffer solution, allowing the transfer buffer to interact with the membrane.
When performing fluorescent western blotting, standard PVDF membranes commonly exhibit high levels of autofluorescence, as can nitrocellulose membranes, particularly in lower-wavelength channels (i.e., UV, 488 and 550 nm). This is a significant disadvantage because the high level of fluorescent background can interfere with accurate detection and analysis. Therefore, the proper choice of membrane for fluorescent western blotting is crucial as it affects the usable fluorescent channels, detection limits, and overall accuracy.
Invitrogen low fluorescence PVDF membranes exhibit minimal auto-fluorescence across all fluorescent channels and result in reduced background compared to both nitrocellulose and standard PVDF membranes (Figure 1). Additionally, Invitrogen low fluorescence PVDF membranes feature a unique 0.3 µm pore size with high consistency and uniformity across the membrane surface, making them an excellent choice for high-quality fluorescent western blotting applications (Figure 2).
Figure 1. Invitrogen low fluorescence PVDF exhibits lower autofluorescence when compared to nitrocellulose and standard PVDF membranes.Unprocessed membranes were hydrated and imaged together in six different fluorescent channels on a compatible digital western blot imaging system. Within each channel, exposure times and contrast settings were consistent for each membrane.
Figure 2. Invitrogen low fluorescence PVDF has fibrous structure and a more uniform pore size distribution than many standard PVDF membranes. (A) Top and cross-section views of Invitrogen low fluorescence PVDF membrane (left) and standard PVDF (right) as imaged via scanning electron microscopy. (B) Pore size distribution of Invitrogen low fluorescence PVDF membrane (red) compared with standard PVDF (gray). A capillary porometer was used to evaluate 12 mm diameter circles of each respective membrane for average pore size and frequency.
Figure 3.Invitrogen low fluorescence PVDF exhibits lower autofluorescence when compared to other commercially available low fluorescence PVDF membranes. Unprocessed membranes were hydrated and imaged together in six different fluorescent channels on a compatible digital western blot imaging system using exposure times and contrasting consistent with standard western blot applications. Within each channel, contrasts and imaging time were consistent for each membrane. Membranes were hydrated in 100% methanol for 3 minutes and washed in diH2O for 5 minutes on an orbital shaker.
Compared to other commercially available low fluorescence PVDF membranes, Invitrogen low fluorescence PVDF membranes offer exceptional performance when imaging in lower wavelength channels (Figure 3).
Lower background fluorescence allows for more protein targets per blot, higher-quality data publication, and several additional benefits including:
Figure 4. Invitrogen low fluorescence PVDF achieves higher signal-to-noise ratio in western blotting compared to other commercially available low fluorescence PVDF membranes. Four lysates were 2-fold serially diluted from 20 µg–2.5 µg, loaded and electrophoresed on Tris-Glycine gels, and transferred to three different membranes. Blocking was completed for 1 hour at room temperature, followed by overnight 4°C incubation with primary antibodies against Hsp90, Calreticulin, beta-Actin, and p23. Secondary antibodies (GAR-800, GAM-647, and GAC-488) in TBST were applied for 1 hour at room temperature. Blots were imaged and contrasted on the iBright FL1500 Imaging System (Cat. No. A44241) under identical conditions.
Western blotting filter papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels (SDS-PAGE) to PVDF, nitrocellulose, or other membranes. Sheets of filter paper are necessary components of transfer sandwiches and cassettes that typically must be assembled for various kinds of protein or nucleic acid, gel-to-membrane, transfer protocols. High-quality western blotting filter papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels and midi gels. Choose a paper with the thickness and dimensions appropriate for specific gel sizes and device platforms or cassettes.
Thickness | Mini Size | Midi Size | Special Sizes |
---|---|---|---|
0.83 mm | Western Blotting Filter Paper, 7 x 8.4 cm (Cat. No. 84783) | Western Blotting Filter Paper, 8 x 13.5 cm (Cat. No. 84784) | Western Blotting Filter Papers, 8 x 10.5 cm (Cat. No. 88600) |
2.5 mm | Western Blotting Filter Paper, Extra Thick, 7 x 8.4 cm (Cat. No. 88605) Blotting Filter Papers, 2.5 mm thick, 7.5 x 8.4 cm (Cat. No. LC2010) | Western Blotting Filter Paper, Extra Thick, 8 x 13.5 cm (Cat. No. 88615) Blotting Filter Papers, 2.5 mm thick, 8.6 x 13.5 cm (Cat. No. LC2008) | Western Blotting Filter Paper, Extra Thick, 8.5 x 9 cm (Cat. No. 88610) Western Blotting Filter Paper, Extra Thick, 20 x 20 cm (Cat. No. 88620) |
The iBlot 3 Western Blot Transfer System combines an innovative, robust design that helps to deliver exceptional performance and convenience with proven iBlot technology.
The Power Blotter is our flexible solution for western blot transfer, from interchangeable blotting cassettes to suit your required throughput, to multiple transfer stack consumable choices.
The Mini Blot Module is designed to make western transfers easy with a universal connection and molded gasket. The inner core requires less methanol-based transfer buffer per transfer.
Two SureLock Tandem Midi Blot Modules fit into the innovative, dual-purpose SureLock Tandem Midi Gel Tank and are designed for easy, room-temperature gel transfers.
For Research Use Only. Not for use in diagnostic procedures.