5 Steps to Multiplexed Fluorescent Western Blotting
Multiplex fluorescent western blotting provides accurate, quantitative results, stable signals, and the ability to clearly evaluate multiple protein targets on a single blot, which makes this technique increasingly popular. With a range of fluorescent dyes and antibodies for western blot detection, as well as the availability of new imaging systems, multiplexed western blotting can save time, reduce cost, and improve efficiency of data generation and collection.
Here, we divide multiplexed fluorescent western blotting into 5 steps from sample preparation to data collection to help you optimize each step, and avoid pitfalls, in order to achieve your best results.
Follow these 5 steps to multiplexed fluorescent western blots
Sample preparation for protein gel electrophoresis
Protein samples used for western blotting can be obtained from many sources, including tissues, cells, and subcellular fractions. Many methods are available for isolating and purifying proteins from these sources.
It is important when preparing protein samples for fluorescent western blotting experiments to avoid to introduction of contaminants that will contribute to background fluorescence. Bromophenol blue will contribute to background fluorescence during imaging.
Total protein extraction reagents and kits provide extracts that are compatible with western blotting are shown below:
Sample typeGoalRecommended Thermo Scientific reagents or kits
Primary cultured or mammalian cells or tissuesTotal protein extraction
M-PER Reagent
T-PER Reagent
N-PER Reagent
RIPA Lysis and Extraction Buffer
Pierce IP Lysis Buffer
Cultured mammalian cells or tissuesSubcellular fractionation or organelle isolation
NE-PER Reagent
Subcellular Fractionation Kits
Mitochondria Isolation Kits
Pierce Cell Surface Protein Isolation Kit
Syn-PER Reagent
Lysosomes Enrichment Kit
Bacterial cellsTotal protein extractionB-PER Reagent
Yeast cellsTotal protein extraction
Y-PER Reagent
Y-PER Plus Reagent
Insect cells (baculovirus)Total protein extraction
I-PER Reagent
Plant tissue (leaf, stem, root, flower)Total protein extraction
P-PER Reagent
Find related products
Protein quantitation
Knowing the concentration of your protein sample will help you determine the proper amount to load on a gel during the electrophoresis step. A wide variety of protein assays is available for measuring the protein concentration of your sample. Use the Protein Assay Selection Guide to help you determine the right protein assay for your needs.
In addition to reliable protein quantification and analysis, perform UV-Vis photometric research applications such as DNA and RNA analysis and ELISAs with the Thermo Scientific Multiskan Sky Microplate Spectrophotometer. The Multiskan Sky reader features a broad wavelength range (200-1000 nm), path length correction and a fast reading speed. The optional µDrop plate enables microvolume analysis—it's like performing up to 16 NanoDrop measurements at once! The intuitive touchscreen user interface, on-board software and built-in protocols let you run quick measurements directly from the instrument. Alternatively, with any instrument purchase you can use our unlimited license, easy-to use Thermo Scientific SkanIt Software with access to our extensive online library of ready-made protocols.
Separate proteins based on molecular weight using gel electrophoresis
Separation of prepared protein samples using protein gel electrophoresis is the second step of multiplexed fluorescent western blotting. Obtaining good separation of the target proteins from one another can be achieved by careful selection of protein gel chemistry and polyacrylamide concentration. Consider using gradient gels, which provide a polyacrylamide concentration gradient that helps provide sharper banding patterns.
Choose the right protein gel
Obtaining optimal resolution of your target protein can be achieved by choosing the right of protein gel chemistry for your protein. Our precast protein gels are offered in four different chemistries. The choice of whether to use one chemistry or another depends on the abundance of the protein you’re separating, the size of the protein.
Find the right gel for your protein
Convert from other suppliers' gels
Find the right protein gel and buffers for optimal resolution of your protein.
Protein sample typeGel chemistryPrecast gelSample buffer*Protein laddersRunning bufferTransfer buffer
Low-abundance/ posttranslationally modified
Broad-range MW (6–400 kDa)
Bis-TrisBolt Bis-Tris Plus (load up to 60 μL)Fluorescent Compatible Sample BufferiBright Prestained Protein Ladder
Bolt MOPS SDS Buffer (15–260 kDa); Bolt MES SDS Buffer (3.5–160 kDa); add Bolt Antioxidant for reduced samplesBolt Transfer Buffer
NuPAGE Bis-TrisNuPAGE MOPS SDS Buffer (15–260 kDa); Bolt MES SDS Buffer (3.5–160 kDa); add Bolt Antioxidant for reduced samplesNuPAGE Transfer Buffer
High-abundance
Broad-range MW (6–400 kDa)
Tris-glycineNovex Tris-Glycine WedgeWell formatNovex Tris-Glycine SDS Running BufferNovex Tris-Glycine Transfer Buffer
High MW (40–500 kDa)Tris-acetateNuPAGE Tris-Acetate GelsSpectra Multicolor High Range Protein LadderNuPAGE Tris-Acetate SDS Running BufferNuPAGE Transfer Buffer
Low MW (2.5–40 kDa)TricineNovex Tricine Mini GelsSpectra Multicolor Low Range Protein LadderNovex Tricing SDS Running BufferNovex Tris-Glycine Transfer Buffer
*Heat samples at 70°C for 10 min.
Sample Buffers
Sample buffers containing bromophenol blue will fluoresce and can contribute to increased background. If using sample buffers with bromophenol blue, the dye front may be run off the gel prior to transfer or cut from the membrane after transfer to avoid background fluorescence signal.
Consider using fluorescence-compatible sample buffers without bromophenol blue, such as Invitrogen Fluorescent Compatible Sample Buffer (Cat. No. LC2570).
Protein ladders
iBright Prestained Protein Ladder is recommended for use as a molecular weight marker for multiplexed fluorescent western blotting of medium–range molecular weight proteins. It allows direct visualization of protein bands during electrophoresis and features:
iBright Prestained Protein Ladder as it appears in visible light and with chemiluminescent and fluorescent detection methods
  • Ten blue-stained proteins, also fluorophore labeled to enable direct and near-IR fluorescent visualization
  • Two unstained proteins (30 kDa and 80 kDa) that enable direct visualization of chemiluminescent or fluorescent detection
Standard prestained molecular weight markers can be used, but the loading amount will need to be optimized if the marker contains fluorescent bands since overloading can increase background fluorescence and signal bleed-through to adjacent lanes. iBright Prestained Protein Ladder allows you to decrease the amount of molecular weight markers loaded onto the gel: typically, 2–4 μL is sufficient for visualization and fluorescence detection.
Learn more about the iBright Prestained Protein Ladder
Protein gels welcome packs
iBright Prestained Protein Ladder as it appears in visible light and with chemiluminescent and fluorescent detection methods
Protein gels welcome packs are a cost effective way to start using Invitrogen precast protein gels for multiplexed fluorescent western blotting experiments. Protein gels welcome packs are available for each of the protein gel chemistries and come with precast protein mini or midi gels, buffers, ladders, and a Mini Gel Tank or SureLock Tandem Midi Gel Tank to provide you with great savings over the cost of the individual components. iBright Prestained Protein Ladder and Invitrogen Fluorescent Compatible Sample Buffer (Cat. No. LC2570) are not included with protein gels welcome packs and must be ordered separately.
Learn more about the protein gels welcome packs
Transfer the protein from the gel matrix to a solid-support membrane
The efficient transfer of proteins from the polyacrylamide gel after electrophoresis to a nitrocellulose or polyvinylidene difluoride (PVDF) membrane is an important step in western blotting so that specific proteins can be detected using immunodetection techniques.
Electrophoretic methods of gel transfer can be sorted into wet transfer, semi-dry transfer, and dry transfer.
Use the selection table below to compare transfer methods and needed equipment, buffers, and time requirements.
 Wet transferSemi-dry transferDry transfer
Mini Blot Module
XCell II Blot Module
SureLock Tandem
Midi Blot Module
Power Blotter
Systems
iBlot 3 device
iBlot 3 Western Blot Transfer System
Capacity1 mini gel per blot module; 1–2 blot modules per tankUp to 2 mini blots1 midi gel per blot module; 1–2 blot modules per tank1–4 mini or 1–2 midi gels1–2 midi or 1–4 mini gel
Transfer time60 min60–120 min30 min7–10 minas few as 3 min
Blotting area (cm)9 x 99 x 99.2 x 14.410 x 18 or
21 x 22.5
8.5 x 13.5
Transfer buffer volume220 mL per blot module200mL300 mL per blot module
Pre-cut membranes & filters: 50–100 mL;
Select transfer stacks: buffer not required
Buffer not required
Power supplyExternalExternalExternalInternalInternal
Required equipmentMini Gel Tank: capacity for up to 2 Mini Blot ModulesXCell SureLock Mini-CellSureLock Tandem Midi Gel Tank
 Learn moreLearn moreLearn moreLearn moreLearn more
Video: See how easy it is to do a protein gel transfer with the iBlot 3 Western Blot Transfer System.
Video Player is loading.
Current Time 0:00
Duration 5:13
Loaded: 3.14%
Stream Type LIVE
Remaining Time 5:13
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Transfer membranes
    To eliminate a major source of background fluorescence, use membranes with low autofluorescence, including nitrocellulose and specialty low-fluorescence PVDF membranes such as Thermo Scientific Nitrocellulose Membrane (Cat. No. 88018) and Low-Fluorescence PVDF Transfer Membrane (Cat. No. 22860). Only handle membranes with gloved hands and clean blunt forceps to limit contamination and scratches on the membranes, which can contribute to background fluorescence and artifacts.
    Tip: Avoid using pens on membranes, as many inks fluoresce. Use a pencil instead.
    Transfer buffers
    Use the table below to identify the correct transfer buffer for your protein gel.
    Protein sample typeGel chemistryPrecast gelTransfer buffer
    Low-abundance/ posttranslationally modified
    Broad-range MW (6–400 kDa)
    Bis-TrisBolt Bis-Tris Plus (load up to 60 μL)Bolt Transfer Buffer
    NuPAGE Bis-TrisNuPAGE Transfer Buffer
    High-abundance
    Broad-range MW (6–400 kDa)
    Tris-glycineNovex Tris-Glycine WedgeWell formatNovex Tris-Glycine Transfer Buffer
    High MW (40–500 kDa)Tris-acetateNuPAGE Tris-Acetate GelsNuPAGE Transfer Buffer
    Low MW (2.5–40 kDa)TricineNovex Tricine Mini GelsNovex Tris-Glycine Transfer Buffer
    Detect transferred proteins with fluorescently labelled antibodies
    Careful selection of antibodies and fluorescent labels is required for successful multiplexed fluorescent western blotting. Download our two-part app note: Fluorescent western blotting two-part app note—from basic technique to multiplexing for a full discussion of antibody and fluorescent label selection.
    Choosing fluorescent labels
    Select fluorophores with optically distinct spectra to avoid cross-channel fluorescence. Examples of fluorophores chosen for multiplex experiments for distinct excitation spectra and emission spectra are shown in figures 1 and 2, below. See Table 1 for examples of multiplex fluorophore combinations that can be used with the iBright FL1500 Imaging System for 1- to 4-probe multiplexed western experiments.
    Table 1. Examples of fluorophore combinations for successful multiplexing with the iBright FL Imaging System.
    Number of targetsConjugate 1Conjugate 2Conjugate 3Conjugate 4
    1Alexa Fluor Plus 647   
    2Alexa Fluor Plus 647Alexa Fluor 546  
    3Alexa Fluor Plus 647Alexa Fluor 546Alexa Fluor Plus 488 
    4Alexa Fluor Plus 647Alexa Fluor 546Alexa Fluor Plus 488Alexa Fluor Plus 800
    Example of multiplex experiment with carefully chosen fluorophores with distinct excitation spectra
    Figure 1. Example of multiplex experiment with carefully chosen fluorophores with distinct excitation spectra. In this example generated on the Fluorescence SpectraViewer, the excitation spectra (dashed lines) of Alexa Fluor Plus 488 and Alexa Fluor 546 fluorophores have minimal overlap within the range of the excitation filter. Despite both fluorophores having part of their emission spectra (solid lines) within the range of the emission filter, Alexa Fluor 546 would not be excited by the excitation filter that has been selected for Alexa Fluor Plus 488, so no fluorescence from Alexa Fluor 546 would be present to go through the emission filter.
    Example of multiplex experiment with carefully chosen fluorophores with distinct emission spectra
    Figure 2. Example of multiplex experiment with carefully chosen fluorophores with distinct emission spectra. In this example generated on the Fluorescence SpectraViewer, the emission spectra (solid lines) of Alexa Fluor Plus 680 and Alexa Fluor 790 have no overlap within the ranges of the two emission filters. Despite both fluorophores having part of their excitation spectra (dashed lines) within the range of excitation filter 1, any Alexa Fluor 790 fluorescence generated by that excitation range is not within the wavelengths allowed to pass through emission filter 1, so no fluorescence from Alexa Fluor 790 would reach the camera detector in that channel.
    Alexa Fluor Plus Secondary Antibodies
    Alexa Fluor Plus Secondary Antibodies
    Need to detect and visualize low-abundance targets in rare or precious samples by fluorescent western blot?
    Compare to traditional Alexa Fluor secondary antibodies:
    • Higher signal-to-noise ratio
    • Lower cross-reactivity
    Learn more
    Search for specific antibodies
    Find antibodies of interest using the search tool below. Then filter the results by target or host species, monoclonal or polyclonal antibody type, application, and other criteria.
    Search now
    Find related products
    Automation of the detection process
    ibind
    There is a hands-free alternative for all blocking, antibody incubation, and washing steps using our Invitrogen iBind and iBind Flex Western Devices. Find out more at thermofisher.com/ibind.
    Troubleshooting
    ProblemPossible causeSolution
    Weak or no signalInsufficient amount of primary antibody
    • Increase primary antibody concentration
    • Ensure primary antibody has a good titer and is specific for the antigen to be detected
    • Extend the incubation time to overnight at 4°C, or 3–6 hours at room temperature
    • Try using an antibody enhancer
    Lost activity of antibody
    • Ensure the antibody was stored appropriately
    • Check the expiration date of the antibody
    • Avoid multiple uses of pre-diluted antibodies
    Imaging exposure time is too short
    • Increase exposure time
    • Use the Smart Exposure feature on the iBright FL1500 system, or a comparable autoexposure feature on another instrument
    Incorrect instrument settings
    • Ensure the correct excitation and emission ranges are selected for the intended fluorophore
    Use of detergent
    • Too much detergent or the nature of the detergent can result in washing away the signal—decrease or eliminate detergent
    Blocking buffer blocks antigen
    • Some blocking solutions can mask the blot and reduce the availability of the antigen to the antibody, especially if the blocking step is >1 hour
    • Dilute your primary antibody in wash buffer
    • Evaluate another blocking buffer
    Insufficient quantity of sample loaded on the gel
    • Too little lysate leads to insufficient availability of the target of interest
    • Perform serial dilutions of the lysate or sample to determine the optimal amount of protein to load
    Poor transfer of protein or loss of the protein after transfer
    • Check transfer conditions to confirm protein transfer
    • Reoptimization may be required when probing for a new protein
    Nonspecific bandsPoor antibody specificity for the target of interest
    Poor sample integrity
    • Sample degradation due to overheating or protease activity results in target breakdown and low target recognition by the antibody
    Antibody cross-reactivity in multiplex detection
    • Choose primary antibodies raised in distantly related species
    • Use highly cross-adsorbed secondary antibodies
    • Reduce the amount of the secondary antibody used to remain within the optimal performance range
    Fluorescent bleed-through from another channel when multiplexing (appearance of an unexpected band)
    • Usa a tool like Fluorescence SpectraViewer to visualize fluorphore spectra, and avoid spectrally close conjugates, especially when the signal is very strong
    • Ensure that your fluorescent dyes can be distinctively detected on your imager
    • Use the autoexposure feature on the instrument to determine the optimal exposure time for each channel
    Background issues (high, uneven, or speckled)High background due to membrane contamination
    • Handle the membrane carefully using clean dishes or trays and clean forceps
    • Determine the best blocking buffer for your application—primary antibodies will react differently in different blocking buffers; blocking buffers like normal animal sera or milk may result in cross-reactivity
    Artifacts from overloading the protein marker or ladder
    • Load less of the molecular weight marker onto the gel
    Nonoptimal wash or diluent solutions
    • Use a wash buffer with 0.1–0.2% Tween 20 detergent
    • Prepare the secondary antibody diluent with 0.05% Tween 20 detergent
    • Increase the number or duration of wash steps
    High background from an excess of secondary antibody
    • Optimize the secondary antibody dilution depending on the dye being used, following the vendor's recommended dilution and adapting accordingly
    Blotchy or uneven background due to the membrane drying out
    • Ensure good coverage of the whole blot during all incubation steps
    • Ensure consistent agitation during every incubation step
    Incorrect choice of membrane
    • The membrane type can affect the background; for example, PVDF membranes can autofluoresce and cause high background, so use low-fluorescence PVDF membranes
    Dust and fingerprints on the membrane
    • Use clean forceps to handle the membrane, and avoid directly touching the membrane; particulates and contaminants from unclean tools may fluoresce
    • Use clean incubation trays or dishes—rinsing with methanol followed by water will help dissolve residual dried dyes from previous uses
    • Clean transfer devices and dusty consumables (e.g., pads) if using a wet transfer method as they can introduce speckles
    • Clean the imager surface with ethanol to remove dust, lint, and residues before placing a blot for imaging
    Imaging of the fluorescent western blot and quantitation of proteins
    With the latest advances in imaging software and instrument sensitivity, fluorescent image capture and quantitative western blot analysis is now easier. Be certain the instrument you plan to use is capable of capturing the number of fluorophores you would like to detect and whether the imager has the correct filters for use with the fluorophores.
    We offer iBright FL1500 Imaging System, a powerful, easy-to-use system, which provides sensitive, streamlined, multimode image capture (see image 2 below). The iBright FL1500 is capable of easily capturing 4-plex images. It features a large capacitive touch-screen interface and intelligently designed software.
    See product details for the iBright FL1500 Imaging System
    Download the iBright FL1500 Imaging System brochure
    Four-channel imaging of a multiplexed fluorescent western blot
    Figure 2. Four-channel imaging of a multiplexed fluorescent western blot.Up to four different proteins can be imaged simultaneously on the same blot. HA-tagged RB-1 was expressed in HeLa cell extract using the 1-Step Human High-Yield Mini IVT Kit (Cat. No. 88890) and appropriate expression-ready clones. The resulting reaction mixture was prepared for reducing SDS-PAGE, serially diluted, and electrophoresed on a Novex WedgeWell 4–20% Tris-glycine gel (Cat. No. XP04200PK2). The protein was transferred to a PVDF membrane using the Pierce Power Blotter (Cat. No. 22834), and the membrane was blocked and probed with the following primary antibodies: chicken anti-calreticulin (Cat. No. PA1-903), rabbit anti-HSP90 (Cat. No. PA3-013), and mouse anti-p23 (Cat. No. MA3-414). The membrane was washed and probed with the following secondary antibodies in TBS-Tween 20: goat anti-chicken Alexa Fluor 546 (Cat. No. A11040) (pseudocolored in yellow), goat anti-rabbit Alexa Fluor Plus 800 (Cat. No. A32735) (pseudocolored in green), and goat anti-mouse Alexa Fluor Plus 680 (Cat. No. A32729) (pseudocolored in red). The membrane was again washed and probed for 1 hr with mouse anti-HA primary antibody directly conjugated to Alexa Fluor 488 (Cat. No. 26183-D488) (pseudocolored in blue), in TBS-Tween 20. The membrane was washed and imaged on the iBright FL1000 Imaging System.
    Quantitation
    Protein normalization is a critical step in obtaining reliable and reproducible quantitative western blotting and can be performed using housekeeping proteins or total protein normalization. Under ideal conditions, normalization would not be necessary, but factors such as sample loading and transfer efficiency make normalizing the western blot essential. Total protein normalization is a more reliable method, as housekeeping proteins can be affected by experimental conditions. For total protein normalization of western blotting data, the Invitrogen No-Stain Protein Labeling Reagent is a fast, easy-to-use reagent applied to a membrane after transfer and provides sensitive, linear detection.
    The iBright FL1500 Imaging System includes software for effortless normalization using the No-Stain Protein Labeling Reagent and many other methods.
    Learn more about the No-Stain Protein Labeling Reagent
    Download this technical note, which provides the basic principles of normalization using internal loading controls and describes how to accurately normalize western blots to obtain meaningful, reproducible data.
    Application note: Normalization in western blotting to achieve relative quantitation
    Reprobing fluorescent western blots
    To reprobe the blot with other antibodies, use Restore Fluorescent Western Blot Stripping Buffer. Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only.
    See product details for Restore Fluorescent Western Blot Stripping Buffer
    iWestern Workflow
    Need a start-to-finish western blotting solution?
    Or just looking to boost the performance of one of the main blotting steps?
    Check out the iWestern workflow
    Sample preparation for protein gel electrophoresis
    Protein samples used for western blotting can be obtained from many sources, including tissues, cells, and subcellular fractions. Many methods are available for isolating and purifying proteins from these sources.
    It is important when preparing protein samples for fluorescent western blotting experiments to avoid to introduction of contaminants that will contribute to background fluorescence. Bromophenol blue will contribute to background fluorescence during imaging.
    Total protein extraction reagents and kits provide extracts that are compatible with western blotting are shown below:
    Sample typeGoalRecommended Thermo Scientific reagents or kits
    Primary cultured or mammalian cells or tissuesTotal protein extraction
    M-PER Reagent
    T-PER Reagent
    N-PER Reagent
    RIPA Lysis and Extraction Buffer
    Pierce IP Lysis Buffer
    Cultured mammalian cells or tissuesSubcellular fractionation or organelle isolation
    NE-PER Reagent
    Subcellular Fractionation Kits
    Mitochondria Isolation Kits
    Pierce Cell Surface Protein Isolation Kit
    Syn-PER Reagent
    Lysosomes Enrichment Kit
    Bacterial cellsTotal protein extractionB-PER Reagent
    Yeast cellsTotal protein extraction
    Y-PER Reagent
    Y-PER Plus Reagent
    Insect cells (baculovirus)Total protein extraction
    I-PER Reagent
    Plant tissue (leaf, stem, root, flower)Total protein extraction
    P-PER Reagent
    Find related products
    Protein quantitation
    Knowing the concentration of your protein sample will help you determine the proper amount to load on a gel during the electrophoresis step. A wide variety of protein assays is available for measuring the protein concentration of your sample. Use the Protein Assay Selection Guide to help you determine the right protein assay for your needs.
    In addition to reliable protein quantification and analysis, perform UV-Vis photometric research applications such as DNA and RNA analysis and ELISAs with the Thermo Scientific Multiskan Sky Microplate Spectrophotometer. The Multiskan Sky reader features a broad wavelength range (200-1000 nm), path length correction and a fast reading speed. The optional µDrop plate enables microvolume analysis—it's like performing up to 16 NanoDrop measurements at once! The intuitive touchscreen user interface, on-board software and built-in protocols let you run quick measurements directly from the instrument. Alternatively, with any instrument purchase you can use our unlimited license, easy-to use Thermo Scientific SkanIt Software with access to our extensive online library of ready-made protocols.
    Separate proteins based on molecular weight using gel electrophoresis
    Separation of prepared protein samples using protein gel electrophoresis is the second step of multiplexed fluorescent western blotting. Obtaining good separation of the target proteins from one another can be achieved by careful selection of protein gel chemistry and polyacrylamide concentration. Consider using gradient gels, which provide a polyacrylamide concentration gradient that helps provide sharper banding patterns.
    Choose the right protein gel
    Obtaining optimal resolution of your target protein can be achieved by choosing the right of protein gel chemistry for your protein. Our precast protein gels are offered in four different chemistries. The choice of whether to use one chemistry or another depends on the abundance of the protein you’re separating, the size of the protein.
    Find the right gel for your protein
    Convert from other suppliers' gels
    Find the right protein gel and buffers for optimal resolution of your protein.
    Protein sample typeGel chemistryPrecast gelSample buffer*Protein laddersRunning bufferTransfer buffer
    Low-abundance/ posttranslationally modified
    Broad-range MW (6–400 kDa)
    Bis-TrisBolt Bis-Tris Plus (load up to 60 μL)Fluorescent Compatible Sample BufferiBright Prestained Protein Ladder
    Bolt MOPS SDS Buffer (15–260 kDa); Bolt MES SDS Buffer (3.5–160 kDa); add Bolt Antioxidant for reduced samplesBolt Transfer Buffer
    NuPAGE Bis-TrisNuPAGE MOPS SDS Buffer (15–260 kDa); Bolt MES SDS Buffer (3.5–160 kDa); add Bolt Antioxidant for reduced samplesNuPAGE Transfer Buffer
    High-abundance
    Broad-range MW (6–400 kDa)
    Tris-glycineNovex Tris-Glycine WedgeWell formatNovex Tris-Glycine SDS Running BufferNovex Tris-Glycine Transfer Buffer
    High MW (40–500 kDa)Tris-acetateNuPAGE Tris-Acetate GelsSpectra Multicolor High Range Protein LadderNuPAGE Tris-Acetate SDS Running BufferNuPAGE Transfer Buffer
    Low MW (2.5–40 kDa)TricineNovex Tricine Mini GelsSpectra Multicolor Low Range Protein LadderNovex Tricing SDS Running BufferNovex Tris-Glycine Transfer Buffer
    *Heat samples at 70°C for 10 min.
    Sample Buffers
    Sample buffers containing bromophenol blue will fluoresce and can contribute to increased background. If using sample buffers with bromophenol blue, the dye front may be run off the gel prior to transfer or cut from the membrane after transfer to avoid background fluorescence signal.
    Consider using fluorescence-compatible sample buffers without bromophenol blue, such as Invitrogen Fluorescent Compatible Sample Buffer (Cat. No. LC2570).
    Protein ladders
    iBright Prestained Protein Ladder is recommended for use as a molecular weight marker for multiplexed fluorescent western blotting of medium–range molecular weight proteins. It allows direct visualization of protein bands during electrophoresis and features:
    iBright Prestained Protein Ladder as it appears in visible light and with chemiluminescent and fluorescent detection methods
    • Ten blue-stained proteins, also fluorophore labeled to enable direct and near-IR fluorescent visualization
    • Two unstained proteins (30 kDa and 80 kDa) that enable direct visualization of chemiluminescent or fluorescent detection
    Standard prestained molecular weight markers can be used, but the loading amount will need to be optimized if the marker contains fluorescent bands since overloading can increase background fluorescence and signal bleed-through to adjacent lanes. iBright Prestained Protein Ladder allows you to decrease the amount of molecular weight markers loaded onto the gel: typically, 2–4 μL is sufficient for visualization and fluorescence detection.
    Learn more about the iBright Prestained Protein Ladder
    Protein gels welcome packs
    iBright Prestained Protein Ladder as it appears in visible light and with chemiluminescent and fluorescent detection methods
    Protein gels welcome packs are a cost effective way to start using Invitrogen precast protein gels for multiplexed fluorescent western blotting experiments. Protein gels welcome packs are available for each of the protein gel chemistries and come with precast protein mini or midi gels, buffers, ladders, and a Mini Gel Tank or SureLock Tandem Midi Gel Tank to provide you with great savings over the cost of the individual components. iBright Prestained Protein Ladder and Invitrogen Fluorescent Compatible Sample Buffer (Cat. No. LC2570) are not included with protein gels welcome packs and must be ordered separately.
    Learn more about the protein gels welcome packs
    Transfer the protein from the gel matrix to a solid-support membrane
    The efficient transfer of proteins from the polyacrylamide gel after electrophoresis to a nitrocellulose or polyvinylidene difluoride (PVDF) membrane is an important step in western blotting so that specific proteins can be detected using immunodetection techniques.
    Electrophoretic methods of gel transfer can be sorted into wet transfer, semi-dry transfer, and dry transfer.
    Use the selection table below to compare transfer methods and needed equipment, buffers, and time requirements.
     Wet transferSemi-dry transferDry transfer
    Mini Blot Module
    XCell II Blot Module
    SureLock Tandem
    Midi Blot Module
    Power Blotter
    Systems
    iBlot 3 device
    iBlot 3 Western Blot Transfer System
    Capacity1 mini gel per blot module; 1–2 blot modules per tankUp to 2 mini blots1 midi gel per blot module; 1–2 blot modules per tank1–4 mini or 1–2 midi gels1–2 midi or 1–4 mini gel
    Transfer time60 min60–120 min30 min7–10 minas few as 3 min
    Blotting area (cm)9 x 99 x 99.2 x 14.410 x 18 or
    21 x 22.5
    8.5 x 13.5
    Transfer buffer volume220 mL per blot module200mL300 mL per blot module
    Pre-cut membranes & filters: 50–100 mL;
    Select transfer stacks: buffer not required
    Buffer not required
    Power supplyExternalExternalExternalInternalInternal
    Required equipmentMini Gel Tank: capacity for up to 2 Mini Blot ModulesXCell SureLock Mini-CellSureLock Tandem Midi Gel Tank
     Learn moreLearn moreLearn moreLearn moreLearn more
    Video: See how easy it is to do a protein gel transfer with the iBlot 3 Western Blot Transfer System.
    Video Player is loading.
    Current Time 0:00
    Duration 5:13
    Loaded: 3.14%
    Stream Type LIVE
    Remaining Time 5:13
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Transfer membranes
      To eliminate a major source of background fluorescence, use membranes with low autofluorescence, including nitrocellulose and specialty low-fluorescence PVDF membranes such as Thermo Scientific Nitrocellulose Membrane (Cat. No. 88018) and Low-Fluorescence PVDF Transfer Membrane (Cat. No. 22860). Only handle membranes with gloved hands and clean blunt forceps to limit contamination and scratches on the membranes, which can contribute to background fluorescence and artifacts.
      Tip: Avoid using pens on membranes, as many inks fluoresce. Use a pencil instead.
      Transfer buffers
      Use the table below to identify the correct transfer buffer for your protein gel.
      Protein sample typeGel chemistryPrecast gelTransfer buffer
      Low-abundance/ posttranslationally modified
      Broad-range MW (6–400 kDa)
      Bis-TrisBolt Bis-Tris Plus (load up to 60 μL)Bolt Transfer Buffer
      NuPAGE Bis-TrisNuPAGE Transfer Buffer
      High-abundance
      Broad-range MW (6–400 kDa)
      Tris-glycineNovex Tris-Glycine WedgeWell formatNovex Tris-Glycine Transfer Buffer
      High MW (40–500 kDa)Tris-acetateNuPAGE Tris-Acetate GelsNuPAGE Transfer Buffer
      Low MW (2.5–40 kDa)TricineNovex Tricine Mini GelsNovex Tris-Glycine Transfer Buffer
      Detect transferred proteins with fluorescently labelled antibodies
      Careful selection of antibodies and fluorescent labels is required for successful multiplexed fluorescent western blotting. Download our two-part app note: Fluorescent western blotting two-part app note—from basic technique to multiplexing for a full discussion of antibody and fluorescent label selection.
      Choosing fluorescent labels
      Select fluorophores with optically distinct spectra to avoid cross-channel fluorescence. Examples of fluorophores chosen for multiplex experiments for distinct excitation spectra and emission spectra are shown in figures 1 and 2, below. See Table 1 for examples of multiplex fluorophore combinations that can be used with the iBright FL1500 Imaging System for 1- to 4-probe multiplexed western experiments.
      Table 1. Examples of fluorophore combinations for successful multiplexing with the iBright FL Imaging System.
      Number of targetsConjugate 1Conjugate 2Conjugate 3Conjugate 4
      1Alexa Fluor Plus 647   
      2Alexa Fluor Plus 647Alexa Fluor 546  
      3Alexa Fluor Plus 647Alexa Fluor 546Alexa Fluor Plus 488 
      4Alexa Fluor Plus 647Alexa Fluor 546Alexa Fluor Plus 488Alexa Fluor Plus 800
      Example of multiplex experiment with carefully chosen fluorophores with distinct excitation spectra
      Figure 1. Example of multiplex experiment with carefully chosen fluorophores with distinct excitation spectra. In this example generated on the Fluorescence SpectraViewer, the excitation spectra (dashed lines) of Alexa Fluor Plus 488 and Alexa Fluor 546 fluorophores have minimal overlap within the range of the excitation filter. Despite both fluorophores having part of their emission spectra (solid lines) within the range of the emission filter, Alexa Fluor 546 would not be excited by the excitation filter that has been selected for Alexa Fluor Plus 488, so no fluorescence from Alexa Fluor 546 would be present to go through the emission filter.
      Example of multiplex experiment with carefully chosen fluorophores with distinct emission spectra
      Figure 2. Example of multiplex experiment with carefully chosen fluorophores with distinct emission spectra. In this example generated on the Fluorescence SpectraViewer, the emission spectra (solid lines) of Alexa Fluor Plus 680 and Alexa Fluor 790 have no overlap within the ranges of the two emission filters. Despite both fluorophores having part of their excitation spectra (dashed lines) within the range of excitation filter 1, any Alexa Fluor 790 fluorescence generated by that excitation range is not within the wavelengths allowed to pass through emission filter 1, so no fluorescence from Alexa Fluor 790 would reach the camera detector in that channel.
      Alexa Fluor Plus Secondary Antibodies
      Alexa Fluor Plus Secondary Antibodies
      Need to detect and visualize low-abundance targets in rare or precious samples by fluorescent western blot?
      Compare to traditional Alexa Fluor secondary antibodies:
      • Higher signal-to-noise ratio
      • Lower cross-reactivity
      Learn more
      Search for specific antibodies
      Find antibodies of interest using the search tool below. Then filter the results by target or host species, monoclonal or polyclonal antibody type, application, and other criteria.
      Search now
      Find related products
      Automation of the detection process
      ibind
      There is a hands-free alternative for all blocking, antibody incubation, and washing steps using our Invitrogen iBind and iBind Flex Western Devices. Find out more at thermofisher.com/ibind.
      Troubleshooting
      ProblemPossible causeSolution
      Weak or no signalInsufficient amount of primary antibody
      • Increase primary antibody concentration
      • Ensure primary antibody has a good titer and is specific for the antigen to be detected
      • Extend the incubation time to overnight at 4°C, or 3–6 hours at room temperature
      • Try using an antibody enhancer
      Lost activity of antibody
      • Ensure the antibody was stored appropriately
      • Check the expiration date of the antibody
      • Avoid multiple uses of pre-diluted antibodies
      Imaging exposure time is too short
      • Increase exposure time
      • Use the Smart Exposure feature on the iBright FL1500 system, or a comparable autoexposure feature on another instrument
      Incorrect instrument settings
      • Ensure the correct excitation and emission ranges are selected for the intended fluorophore
      Use of detergent
      • Too much detergent or the nature of the detergent can result in washing away the signal—decrease or eliminate detergent
      Blocking buffer blocks antigen
      • Some blocking solutions can mask the blot and reduce the availability of the antigen to the antibody, especially if the blocking step is >1 hour
      • Dilute your primary antibody in wash buffer
      • Evaluate another blocking buffer
      Insufficient quantity of sample loaded on the gel
      • Too little lysate leads to insufficient availability of the target of interest
      • Perform serial dilutions of the lysate or sample to determine the optimal amount of protein to load
      Poor transfer of protein or loss of the protein after transfer
      • Check transfer conditions to confirm protein transfer
      • Reoptimization may be required when probing for a new protein
      Nonspecific bandsPoor antibody specificity for the target of interest
      Poor sample integrity
      • Sample degradation due to overheating or protease activity results in target breakdown and low target recognition by the antibody
      Antibody cross-reactivity in multiplex detection
      • Choose primary antibodies raised in distantly related species
      • Use highly cross-adsorbed secondary antibodies
      • Reduce the amount of the secondary antibody used to remain within the optimal performance range
      Fluorescent bleed-through from another channel when multiplexing (appearance of an unexpected band)
      • Usa a tool like Fluorescence SpectraViewer to visualize fluorphore spectra, and avoid spectrally close conjugates, especially when the signal is very strong
      • Ensure that your fluorescent dyes can be distinctively detected on your imager
      • Use the autoexposure feature on the instrument to determine the optimal exposure time for each channel
      Background issues (high, uneven, or speckled)High background due to membrane contamination
      • Handle the membrane carefully using clean dishes or trays and clean forceps
      • Determine the best blocking buffer for your application—primary antibodies will react differently in different blocking buffers; blocking buffers like normal animal sera or milk may result in cross-reactivity
      Artifacts from overloading the protein marker or ladder
      • Load less of the molecular weight marker onto the gel
      Nonoptimal wash or diluent solutions
      • Use a wash buffer with 0.1–0.2% Tween 20 detergent
      • Prepare the secondary antibody diluent with 0.05% Tween 20 detergent
      • Increase the number or duration of wash steps
      High background from an excess of secondary antibody
      • Optimize the secondary antibody dilution depending on the dye being used, following the vendor's recommended dilution and adapting accordingly
      Blotchy or uneven background due to the membrane drying out
      • Ensure good coverage of the whole blot during all incubation steps
      • Ensure consistent agitation during every incubation step
      Incorrect choice of membrane
      • The membrane type can affect the background; for example, PVDF membranes can autofluoresce and cause high background, so use low-fluorescence PVDF membranes
      Dust and fingerprints on the membrane
      • Use clean forceps to handle the membrane, and avoid directly touching the membrane; particulates and contaminants from unclean tools may fluoresce
      • Use clean incubation trays or dishes—rinsing with methanol followed by water will help dissolve residual dried dyes from previous uses
      • Clean transfer devices and dusty consumables (e.g., pads) if using a wet transfer method as they can introduce speckles
      • Clean the imager surface with ethanol to remove dust, lint, and residues before placing a blot for imaging
      Imaging of the fluorescent western blot and quantitation of proteins
      With the latest advances in imaging software and instrument sensitivity, fluorescent image capture and quantitative western blot analysis is now easier. Be certain the instrument you plan to use is capable of capturing the number of fluorophores you would like to detect and whether the imager has the correct filters for use with the fluorophores.
      We offer iBright FL1500 Imaging System, a powerful, easy-to-use system, which provides sensitive, streamlined, multimode image capture (see image 2 below). The iBright FL1500 is capable of easily capturing 4-plex images. It features a large capacitive touch-screen interface and intelligently designed software.
      See product details for the iBright FL1500 Imaging System
      Download the iBright FL1500 Imaging System brochure
      Four-channel imaging of a multiplexed fluorescent western blot
      Figure 2. Four-channel imaging of a multiplexed fluorescent western blot.Up to four different proteins can be imaged simultaneously on the same blot. HA-tagged RB-1 was expressed in HeLa cell extract using the 1-Step Human High-Yield Mini IVT Kit (Cat. No. 88890) and appropriate expression-ready clones. The resulting reaction mixture was prepared for reducing SDS-PAGE, serially diluted, and electrophoresed on a Novex WedgeWell 4–20% Tris-glycine gel (Cat. No. XP04200PK2). The protein was transferred to a PVDF membrane using the Pierce Power Blotter (Cat. No. 22834), and the membrane was blocked and probed with the following primary antibodies: chicken anti-calreticulin (Cat. No. PA1-903), rabbit anti-HSP90 (Cat. No. PA3-013), and mouse anti-p23 (Cat. No. MA3-414). The membrane was washed and probed with the following secondary antibodies in TBS-Tween 20: goat anti-chicken Alexa Fluor 546 (Cat. No. A11040) (pseudocolored in yellow), goat anti-rabbit Alexa Fluor Plus 800 (Cat. No. A32735) (pseudocolored in green), and goat anti-mouse Alexa Fluor Plus 680 (Cat. No. A32729) (pseudocolored in red). The membrane was again washed and probed for 1 hr with mouse anti-HA primary antibody directly conjugated to Alexa Fluor 488 (Cat. No. 26183-D488) (pseudocolored in blue), in TBS-Tween 20. The membrane was washed and imaged on the iBright FL1000 Imaging System.
      Quantitation
      Protein normalization is a critical step in obtaining reliable and reproducible quantitative western blotting and can be performed using housekeeping proteins or total protein normalization. Under ideal conditions, normalization would not be necessary, but factors such as sample loading and transfer efficiency make normalizing the western blot essential. Total protein normalization is a more reliable method, as housekeeping proteins can be affected by experimental conditions. For total protein normalization of western blotting data, the Invitrogen No-Stain Protein Labeling Reagent is a fast, easy-to-use reagent applied to a membrane after transfer and provides sensitive, linear detection.
      The iBright FL1500 Imaging System includes software for effortless normalization using the No-Stain Protein Labeling Reagent and many other methods.
      Learn more about the No-Stain Protein Labeling Reagent
      Download this technical note, which provides the basic principles of normalization using internal loading controls and describes how to accurately normalize western blots to obtain meaningful, reproducible data.
      Application note: Normalization in western blotting to achieve relative quantitation
      Reprobing fluorescent western blots
      To reprobe the blot with other antibodies, use Restore Fluorescent Western Blot Stripping Buffer. Restore Fluorescent Western Blot Stripping Buffer enables the reuse of PVDF membranes, simplifying the Western blot optimization process and allowing the same blot to be reprobed with different primary antibodies to detect alternative targets. Restore Fluorescent Western Blot Stripping Buffer is for use with low-fluorescence PVDF membrane only.
      See product details for Restore Fluorescent Western Blot Stripping Buffer
      iWestern Workflow
      Need a start-to-finish western blotting solution?
      Or just looking to boost the performance of one of the main blotting steps?
      Check out the iWestern workflow
      For Research Use Only. Not for use in diagnostic procedures.