Focused ion beam scanning electron microscopy
FIB-SEM Instruments
Scientists and engineers in both academia and industry are constantly facing new challenges that require highly localized characterization of a wide range of samples and materials. The ongoing drive to improve the quality of these materials means that structural and compositional information at the nanoscale is frequently necessary. FIB-SEM instruments generate exactly this kind of data by combining the precise sample modification of FIB with the high-resolution imaging of SEM.
DualBeam instruments
Thermo Fisher Scientific is the industry leader in FIB-SEM technology with more than 30 years of experience with DualBeams. The technology's novel ability to reveal subsurface structural detail, by making precise cuts with a FIB and then imaging the exposed surface with a high-resolution SEM, has led to its acceptance by researchers and engineers in a wide variety of applications. With more than 2,000 Thermo Scientific systems installed around the world, our DualBeams continue to lead the market with cutting-edge capabilities built on technical innovation and a deep store of application knowledge gathered over years of collaborative development with our customers.
TriBeam instruments
The new TriBeam systems are our latest FIB-SEM innovation, featuring the addition of a femtosecond laser, which can cut many materials at rates that are orders of magnitude faster than a typical FIB. A large cross-section (hundreds of micrometers) can be created in less than five minutes. Because the laser has a different removal mechanism (ablation versus the ion sputtering of FIB), it can easily process challenging materials, such as non-conductive or ion-beam-sensitive samples. The extremely short duration of the femtosecond laser pulses introduces almost no artifacts such as heat impact, microcracking, melting, or those typical of traditional mechanical polishing. In most cases, the laser-milled surfaces are clean enough for direct SEM imaging and even for surface-sensitive techniques such as electron backscatter diffraction (EBSD) mapping.
We offer a broad product portfolio and advanced automation capabilities for applications such as transmission electron microscopy (TEM) sample preparation, subsurface and 3D characterization, nano-prototyping, and in situ experimentation.
Style Sheet for Fonts
Style Sheet for Instrument Cards Original
TEM sample preparation
TEM sample preparation is considered to be one of the most critical tasks in materials science research. However, it is also one of the most challenging and time-consuming. The latest technological innovations of DualBeam technology, along with our comprehensive software solutions and application expertise, enable fast and easy preparation of site-specific, high-quality S/TEM (scanning/transmission electron microscopy) samples for a wide range of materials. Thermo Scientific AutoTEM Software adds the capability for fully automated, unattended in situ TEM sample preparation, significantly increasing throughput and bringing you expert-level results regardless of your experience. Learn more about TEM sample prep.
3D Structural analysis
When combined with Thermo Scientific Auto Slice & View Software, DualBeam instruments provide 3D insight into sample structure by selectively removing (milling) the material for subsurface characterization. Digital reconstruction generates multi-modal 3D datasets that can consist of a variety of signals, including backscattered electron (BSE) imaging for maximum materials contrast, energy dispersive spectroscopy (EDS) for compositional information, and electron backscatter diffraction (EBSD) for microstructural and crystallographic information.
The SEM capability of DualBeam instruments offers nanoscale details across a wide range of working conditions, from structural information obtained at 30 keV in STEM mode to charge-free, detailed surface information at lower energies. With unique in-lens detectors, DualBeam systems are designed for simultaneous acquisition of angular/energy-selective secondary-electron and BSE data. Fast, accurate, and reproducible results are provided by our unique SEM column design, which features fully automated lens alignments.
Large-volume, millimeter-scale characterization
Thermo Scientific TriBeam systems are (plasma) FIB-SEM instruments combined with a femtosecond laser for ultrafast material removal and millimeter-scale 3D characterization with nanometer resolution. Learn more about TriBeam instruments. 
Nanoprototyping
As technology continues to miniaturize, the demand for nanoscale devices and structures is ever increasing. The challenge of converting the latest ideas and designs into valuable assets only continues to grow, and significant technical and financial barriers need to be overcome for successful prototyping of nanoscale devices.
Standard nanofabrication batch processes involve the combined use of different machines for each pattern layer. These instruments might include a spin coater for resist application, a lithography tool, wet chemistry for resist development, a plasma cleaner, and deposition or etching equipment for pattern transfer. The use of so many distinct tools results in a costly and time-consuming prototyping process. As researchers push nanotechnology towards smaller dimensions, these established procedures and recipes often no longer keep pace with the demands of rapid development.
Thermo Fisher Scientific offers a smart and efficient way of turning nanoscale designs into reality. The nanopatterning capabilities of our FIB-SEM systems can substantially reduce research and development time. Rapid prototyping with the FIB enables functionality testing before the final device layout is established for batch fabrication. Beam-induced deposition of different materials can be combined with FIB milling without the need for additional aligning lithography steps; patterns can be directly added to deposited structures or existing patterns can be modified. The final patterned substrates are immediately available for further processing or characterization. Together, these produce a robust prototyping workflow that enables you to quickly design, create, and inspect micro- and nanoscale functional prototype devices.
Technology
Technology
Industry
Applications
Life Sciences
Materials Science
Semiconductors
Techniques
Life Sciences
Materials Science
Semiconductors
Helios 6 HXS FIB-SEM
  • Automated material handling
  • Automated data connectivity
  • Automated FIB-SEM workflows
Helios 6 HX FIB-SEM
  • Fully auto TEM sample prep
  • Sub-nm targeting accuracy
  • Robust FIB-SEM workflow
Apreo ChemiSEM System
  • Integrated SEM imaging and chemical characterization
  • Enhanced automation to simplify workflows
  • Extended source lifetime and schedulable upgrades
Helios 6 HD FIB-SEM
  • High-volume TEM sample prep
  • Maximized system utilization
  • Failure analysis and metrology
Meridian EX System
  • Novel electron beam probing
  • 10x resolution versus optical
  • <5 nm node defect localization
Hydra Bio Plasma-FIB
  • Volume EM for cryo or resin-embedded samples with 4 switchable ion species
  • Millimeter-scale slicing with high z-precision with Spin Mill Bio Method
  • Integrated light microscope for correlative targeting
Iliad (S)TEM
  • Advanced integration of EELS and (S)TEM optics
  • Electrostatic beam blanker
  • High energy resolution electron source
Krios 5 Cryo-TEM
  • Fast, high-quality data collection
  • High workflow efficiency
  • Increased productivity and automation
  • Designed-in sustainability
Glacios 2 Cryo-TEM
  • High-resolution structure determination tool (<3Å)
  • Unparalleled ease-of-use through AI-enhanced automation
  • Flexible modes of operation standard: SPA, tomography, MicroED
Tundra Cryo-TEM
  • Structural information at biologically relevant resolution
  • Space efficient and cost effective
  • Easy, iterative sample optimization
  • Unique AI algorithms for streamlined data collection
Spectra Ultra TEM
  • New imaging and spectroscopy capabilities on the most beam sensitive materials
  • A leap forward in EDX detection with Ultra-X
  • Column designed to maintain sample integrity
Spectra 300 TEM
  • Highest-resolution structural and chemical information at the atomic level
  • Flexible high-tension range from 30-300 kV
  • Three lens condenser system
Spectra 200 TEM
  • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
  • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
  • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Talos L120C TEM
  • High versatility and stability
  • 4k x 4k Ceta CMOS camera for speed and large FOV
  • TEM magnification range of 25X to 650kX
  • EDS and STEM options for compositional analyses
Metrios AX TEM
  • Automation options to support quality, consistency, metrology, and reduced OPEX
  • Leverages machine learning for superior autofunctions and feature recognition
  • Workflows for both in-situ and ex-situ lamella preparation
Metrios 6 (S)TEM
  • Fully Automating Semiconductor Metrology Workflows
  • Recipe-Free Automation for (S)TEM Metrology
  • Efficient Chemical Analysis
Metrios 6S (S)TEM
  • Fully automated imaging/metrology
  • Automated TEM grid handling
  • Automated data connectivity
ExSolve WTP DualBeam
  • Can prepare site-specific, 20 nm thick lamellae on whole wafers up to 300 mm in diameter
  • Addresses needs requiring automated, high-throughput sampling at advanced technology nodes
Helios 5 EXL DualBeam
  • Low-voltage performance for high sample preparation quality
  • Ultra-high-resolution immersion-lens field-emission SEM column
  • Automated handling of 300 mm FOUP with EFEM (GEM300 compliant)
Helios 5 DualBeam
  • Fully automated, high-quality, ultra-thin TEM sample preparation
  • High throughput, high resolution subsurface and 3D characterization
  • Rapid nanoprototyping capabilities
Helios 5 PFIB DualBeam
  • Gallium-free STEM and TEM sample preparation
  • Multi-modal subsurface and 3D information
  • Next-generation 2.5 μA xenon plasma FIB column
Helios 5 Hydra DualBeam
  • 4 fast switchable ion species (Xe, Ar, O, N) for optimized PFIB processing of a widest range of materials
  • Ga-free TEM sample preparation
  • Extreme high resolution SEM imaging
FIB-SEM and Laser Ablation
  • All three beams have same coincident point for accurate and repeatable cut placement
  • Millimeter-scale cross sections with up to 15,000x faster material removal than a typical FIB
  • Statistically relevant deep subsurface and 3D data analysis
Arctis Cryo-Plasma-FIB
  • Correlation to light microscopy and relocation in TEM
  • High-quality lamellae with consistent thickness
  • Automated high throughput and connectivity for cryo-tomography
Aquilos 2 Cryo-FIB
  • Automation enables production of multiple lamellas
  • Target and extract your structure of interest with lift-out nano-manipulator
  • 3D visualization for high-resolution tomography
Scios 2 DualBeam
  • Full support of magnetic and non-conductive samples
  • High throughput subsurface and 3D characterization
  • Advanced ease of use and automation capabilities
Axia ChemiSEM
  • Live quantitative elemental mapping
  • High fidelity scanning electron microscopy imaging
  • Flexible and easy to use, even for novice users
  • Easy maintenance
Verios 5 XHR SEM
  • Monochromated SEM for sub-nanometer resolution over the full 1 keV to 30 keV energy range
  • Easy access to beam landing energies as low as 20 eV
  • Excellent stability with piezo stage as standard
Quattro ESEM
  • Ultra-versatile high-resolution FEG SEM with unique environmental capability (ESEM)
  • Observe all information from all samples with simultaneous SE and BSE imaging in every mode of operation
Prisma E SEM
  • Entry-level SEM with excellent image quality
  • Easy and quick sample loading and navigation for multiple samples
  • Compatible with a wide range of materials thanks to dedicated vacuum modes
Apreo 2 SEM
  • High-performance SEM for all-round nanometer or sub-nanometer resolution
  • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
  • Excellent performance at long working distance (10 mm)
VolumeScope 2 SEM
  • Isotropic 3D data from large volumes
  • High contrast and resolution in high and low vacuum modes
  • Simple switch between normal SEM use and serial block-face imaging
Phenom Pharos G2 Desktop FEG-SEM
  • FEG source with 1 - 20 kV acceleration voltage range
  • <2.0 nm (SE) and 3.0 nm (BSE) resolution @ 20 kV
  • Optional fully integrated EDS and SE detector
Phenom XL G2 Desktop SEM
  • For large samples (100x100 mm) and ideal for automation
  • <10 nm resolution and up to 200,000x magnification; 4.8 kV up to 20 kV acceleration voltage
  • Optional fully integrated EDS and BSE detector
Phenom ParticleX Battery Desktop SEM
  • Versatile solution for high-quality, in-house analysis
  • Automated system and analysis of multiple samples
  • Testing 10x faster
Phenom ProX G6 Desktop SEM
  • High performance desktop SEM with integrated EDS detector
  • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
  • Optional SE detector
Phenom Pro G6 Desktop SEM
  • High performance desktop SEM
  • Resolution <6 nm (SE) and <8 nm (BSE); magnification up to 350,000x
  • Optional SE detector
Phenom Pure G6 Desktop SEM
  • Entry level desktop SEM
  • Resolution <15 nm; magnification up to 175,000x
  • Longlife CeB6 source
Phenom Perception GSR Desktop SEM
  • Dedicated automated GSR desktop SEM
  • Resolution <10 nm; magnification up to 200,000x
  • Longlife CeB6 source
Phenom ParticleX AM Desktop SEM
  • Versatile desktop SEM with automation software for Additive Manufacturing
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Phenom ParticleX TC Desktop SEM
  • Versatile desktop SEM with automation software for Technical Cleanliness
  • Resolution <10 nm; magnification up to 200,000x
  • Optional SE detector
Phenom ParticleX Steel Desktop SEM
  • SEM and EDS integrated
  • Ease of use
  • Sub-micrometer inclusions
ELITE System
  • Electrical defect localization
  • Thermal mapping
  • Advanced packaging analysis
Hyperion II System
  • Atomic Force Probing
  • Localize transistor faults
  • Integrated PicoCurrent (CAFM)
nProber IV System
  • Localize transistor and BEOL faults
  • Thermal nanoprobing (-40°C to 150°C)
  • Semi-automated operation
Meridian S System
  • Fault Diagnostic with Active Probe Technology
  • Static laser stimulation (SLS / OBIRCH) and photon emission options
  • Supports both micro-probing and probe card device stimulation
Meridian WS-DP System
  • High-sensitivity, low-noise, low-voltage photon emission detection with broadband DBX or InGaAs camera systems
  • Multi-wavelength laser scanning microscope for scan chain analysis, frequency mapping, transistor probing and isolation of faults
Meridian 7 System
  • Dynamic Optical Fault Isolation for 10nm node and below
  • High resolution visible and Infrared light
  • High-yield sample preparation to 5μm widely available
Meridian IV System
  • High sensitivity extended-wavelength DBX photon emission detection
  • Standard InGaAs photon emission detection
  • Laser Scanning Microscope with multiple wavelength options
Centrios Circuit Edit System
  • Superior image / milling resolution
  • Enhanced milling precision and control
  • Built on the Thermo Scientific Helios DualBeam platform
Centrios HX Circuit Edit System
  • Cutting-edge circuit editing for advanced semiconductor nodes with the latest FIB technology
  • Unparalleled milling precision and control with low landing energy to preserve sensitive circuits
  • Advanced navigation and ion beam placement accuracy
MK.4TE ESD and Latch-Up Test System
  • Rapid-relay-based operations—up to 2304 channels
  • Advanced device preconditioning with six separate vector drive levels
  • Fully compliant Latch-Up stimulus and device biasing
Orion3 Electrostatic Discharge Tester
  • Charged device model testing
  • Dual high resolution color cameras
  • Test densities to less than 0.4mm pitch
Celestron Test System
  • Wafer and package level TLP testing
  • High current TLP pulse generator
  • Can be interfaced with semiautomatic probers
  • Intuitive software for control and report generation
Pegasus ESD Test System
  • Testing per the latest industry standards
  • True system level ESD 150pF/330Ω network
  • 2 pin connection via wafer probes to any device
Nexsa
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
K-Alpha
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Vitrobot System
  • Fully Automated Sample vitrification
  • Blotting Device
  • Semi-Automated Grid Transfer
  • High Sample Throughput
CleanMill System
  • Broad ion beam milling and polishing
  • Wide acceleration voltage range
  • Cryogenic sample preparation
  • CleanConnect compatible
Amira Software
  • Support for multi-data/view/channel
  • Interactive high-quality visualization
  • Machine Learning-based segmentation
  • Intuitive recipe creation
Athena Software
  • Ensure traceability of images, data, metadata and experimental workflows
  • Simplify your imaging workflow
  • Improve collaboration
  • Secure and manage data access
Auto Slice & View 5 Software
  • Automated serial sectioning for DualBeam
  • Multi-modal data acquisition (SEM, EDS, EBSD)
  • On-the-fly editing capabilities
AutoScript 4 Software
  • improved reproducibility and accuracy
  • Unattended, high throughput imaging and patterning
  • Supported by Python 3.5-based scripting environment
AutoScript TEM Software
  • Provides a direct link between research needs and microscope automation
  • Enables improved reproducibility and accuracy
  • Focuses time on the microscope for higher throughput
AutoTEM 5 Software
  • Fully automated in situ S/TEM sample preparation
  • Support of top-down, planar and inverted geometry
  • Highly configurable workflow
  • Easy to use, intuitive user interface
Avizo Software
  • Support for multi-data/multi-view, multi-channel, time series, very large data
  • Advanced multi-mode 2D/3D automatic registration
  • Artifact reduction algorithms
CryoFlow Software
  • Secure data sharing for remote and collaborative projects
  • Efficient data management and connectivity for cryo-EM workflows
  • Interactive high-quality visualization for single particle analysis and cryo-electron tomography results
Smart EPU Software
  • Microscope-embedded solution for single particle data acquisition
  • Supports fully autonomous screening
  • Real time image analysis with AI-driven algorithms
Embedded CryoSPARC Live
  • Real-time, multidimensional feedback on sample quality and instrument settings
  • Adjust imaging and processing based on real-time information
  • Fully integrated with Smart EPU Software
iFast Software
  • Macro recorder for faster recipe creating
  • Runner for unattended overnight operation
  • Alignment tools: Image recognition and edge finding
Inspect 3D Software
  • Image processing tools and filters for cross-correlation
  • Feature tracking for image alignment
  • Algebraic reconstruction technique for iterative projection comparison
Maps Software
  • Acquire high-resolution images over large areas
  • Easily find regions of interest
  • Automate image acquisition process
NEXS Software
  • Auto-syncs position/magnification between NEXS and Circuit Edit system for a more seamless user experience
  • Connects to most Thermo Scientific tools used for EFA, PFA and Circuit Edit space
Pergeos Software
Rocks and Minerals
  • Visualize, process, and analyze rock imagery data
  • Perform multi-scale imaging analysis from pore to core
  • Calculate geological and petrophysical properties
Tomography 5 Software
  • Automated solution for 3D Cryo-EM tomography data acquisition
  • Automatic cassette mapping
  • Dose symmetric tilt scheme for optimal electron dose
Velox Software
  • Focus on getting your insights with all-in-one software
  • Interactive live graphical detector layout
  • Time resolved imaging and analytics – for best results in all experiments
3D Reconstruction
  • Intuitive user interface, maximum employability
  • Intuitive fully automated user interface
  • Based on 'shape from shading' technology, no stage tilt required
Elemental Mapping
  • Fast and reliable information on the distribution of elements within the sample or the selected line
  • Easily exported and reported results
FiberMetric
  • Save time by automated measurements
  • Fast and automated collection of all statistical data
  • View and measure micro and nano fibers with unmatched accuracy
Nanobuilder
  • CAD-based prototyping
  • Fully automated job execution, stage navigation, milling, and deposition
  • Automated alignment and drift control
ParticleMetric
  • Integrated software for online and offline analysis
  • Correlating particle features such as diameter, circularity, aspect ratio and convexity
  • Creating image datasets with Automated Image Mapping
Phenom Programming Interface
  • Customize your SEM to fit your workflow
  • Increase efficiency and save time with automated processes
  • Control imaging settings and stage navigation
PoroMetric
  • Correlate pore features such as area, aspect ratio, major and minor axis
  • Acquire images directly from the Desktop SEM
  • Statistical data with high-quality images
ProSuite
  • Automated collection of images
  • Real-time remote control
  • Standard applications included: Automated Image Mapping + Remote User Interface
Quartz PCI/CFR
  • SEM imaging traceability compliant with 21 CFR Part 11
  • Compatible with the Phenom XL and Phenom Pro desktop SEMs
  • Windows 10 64-bit operating system support
μHeater
  • Ultra-fast heating solution for in situ high resolution imaging
  • Fully integrated
  • Temperatures up to 1200 °C
μPolisher
  • Potential to enable large number of novel, unexplored applications
  • Very low energy milling
  • Small spot size for precise local surface treatment
Selectris and Selectris X Imaging Filters
  • Designed for high stability and atomic resolution imaging
  • Straightforward operation
  • Paired with the latest generation Thermo Scientific Falcon 4 Direct Electron Detector
Ceta D Camera
  • Optimum performance at any high tension (20–300 kV)
  • Compatible with post-column filters and spectrometers
  • Movie acquisition for dynamic studies
Falcon 4i Detector
  • High thorughput for more images per hour
  • Unsurpassed imaging quality with high DQE
  • Lossless data compression with EER
Charge Reduction Sample Holder
  • Up to 8 times higher magnification
  • Faster sample preparation
  • Non conductive samples can be imaged in their natural state
Electrical Feedthrough Sample Holder
  • Connect electrical probes to the sample for in situ measurements
  • Sample can be height adjusted from 0–25 mm manually
  • Measurements of probe currents
Eucentric Sample Holder
  • Eucentric tilting and compucentric rotation on a desktop SEM
  • Fast time-to-image with sample loading < 1 minute
  • Real-time 3D sample visualization module
Filter Inserts
  • Filter residue analysis and asbestos analysis
  • Available in two models which support 47 mm (1.85 inch) and 25 mm (1 inch) filters
  • Use on Phenom Desktop SEM
Metallurgical Sample Holder
  • Designed to support resin-mounted samples
  • Preferred solution for metallurgy and when working with inserts
  • Sample size up to 32 mm diameter and 30 mm height
Micro Tool Sample Holder
  • Quick and fast clamping
  • Tilting and rotation allow for easy sample positioning
  • No extra tooling required for sample loading
Motorized Tilt & Rotation Sample Holder
  • Tilt range -10° to +45°
  • Continuous 360° compucentric rotation
  • Controlled by dedicated Motion Control ProSuite application
Nebula Particle Disperser
  • Standard method for uniform dry powder dispersion
  • Avoids particle clusters
  • Used with Phenom Desktop SEM
Resin Mount Inserts
  • A unique sample holder concept
  • Available in 3 models for supporting standard sized samples of 25 mm (~1 inch), 32 mm (~1 ¼ inch) and 40 mm (~1 ½ inch) diameter
Standard Sample Holder
  • Compact stage allowing analysis of samples of up to 100 mm x 100 mm
  • Can be extended with 3 types of resin or metallurgical mount inserts
  • Used with Phenom Desktop SEM
Sample Holder Inserts
  • Faster cross-sectional imaging of coatings and multilayer samples
  • Easy clamping without the need for screws or extra tools
  • Eliminates the need for screws and tools to clamp the sample
 Download datasheet
Tensile Sample Holder
  • Determine batch quality
  • Determine manufacturing consistencys
  • Aid the design process
Temperature Controlled Sample Holder
  • Temperature range -25 °C to +50 °C
  • Temperature accuracy ±1.5 °C
  • Temperature display resolution 0.1 °C
Nanoscale Materials Characterization
Understanding the nanoscale characteristics of materials is the first step in creating the next generation of lighter, stronger, and more energy efficient solutions. With our electron microscopy solutions, we help you simplify this complex material journey and accelerate your nanoscale materials research. Nanoscale workflow is a journey of material from bulk to nanometer scales using our DualBeam FIB-SEM for sample preparation and Transmission Electron Microscopes for material analysis. 
Electron microscopy learning center
Electron microscopy resources and reference materials for microscopy novices, experts, and everyone in between. The electron microscopy learning center provides a variety of informational and educational resources on electron microscopy for students, educators, or anyone that simply wants to learn more about this fascinating technology.
Resources
Style Sheet to change H2 style to p with em-h2-header class
Video Player is loading.
Current Time 0:00
Duration 0:44
Loaded: 0%
Stream Type LIVE
Remaining Time 0:44
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • default, selected
    3D reconstruction of an automotive oil filter casing, acquired with the Helios Hydra DualBeam and Auto Slice & View 4 Software for automated serial sectioning. Horizontal field width = 350 µm.
    Video Player is loading.
    Current Time 0:00
    Duration 1:24
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:24
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • default, selected
      Video Player is loading.
      Current Time 0:00
      Duration 0:44
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:44
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • default, selected
        3D reconstruction of an automotive oil filter casing, acquired with the Helios Hydra DualBeam and Auto Slice & View 4 Software for automated serial sectioning. Horizontal field width = 350 µm.
        Video Player is loading.
        Current Time 0:00
        Duration 1:24
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 1:24
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          Style Sheet for Komodo Tabs
          Contact us
          Electron microscopy services
          To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
          Learn more ›
          Style Sheet for Support and Service footer
          Style Sheet for Fonts
          Style Sheet for Cards