1x1 image pixel for data collection
Gibco cell culture media for healthy mammalian cell cultures
Cell culture medium is vital for providing the necessary components to support cell growth and function. Components in cell culture media supply your culture environment with nutrients and growth factors for the cells to behave as expected. In addition, media can be formulated to regulate environmental conditions in cell culture, including osmotic pressure and pH. A variety of cell culture media formulations have been developed to support a wide range of cell types and experimental applications.
Time-tested and trusted, our Gibco cell culture media line includes products designed to support the growth and maintenance of a variety of mammalian cells and cell lines. We offer ready-to-use cell culture media as well as powdered and concentrated liquid formulations to fit your experimental setup and budget. For your scale-up needs, Gibco Cell Culture for Bioprocessing provides end-to-end products, process development and customization services to help maximize productivity while reducing costs and time to market.
Explore classical media
DMEM
Dulbecco’s Modified Eagle Medium (DMEM), a widely used cell culture medium, is more nutrient-rich than MEM and is proven for use with many mammalian cell types.
RPMI 1640
Roswell Park Memorial Institute (RPMI) 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins.
DMEM F12
Dulbecco's Modified Eagle Medium Nutrient Mixture F-12 (DMEM F12), a 1:1 mixture combining the benefits of DMEM with the added nutrients found in Ham's F-12.
MEM
Minimum Essential Medium (MEM), a commonly used cell culture media, can be modified to support a variety of mammalian cell culture types and applications.
Selecting the right cell culture media
Use the Gibco Media selection tool to choose from classical basal media and innovative formulations.
Innovative cell culture formulations
We are committed to innovating Gibco cell culture media. We go beyond basic formulations to provide optimized and innovative cell culture solutions in support of your efforts to achieve consistently reproducible results.
BenchStable Media
Gibco BenchStable Media, available in our most common cell culture media formulations, are stable at room temperature, allowing for convenient and flexible storage without utilizing cold storage space.
Human Plasma-Like Medium
Gibco Human Plasma-Like Medium (HPLM) is formulated to mimic the metabolic profile of human plasma, allowing for improved physiological relevance when studying cancer and disease.
Media with GlutaMAX Supplement
Media with Gibco GlutaMAX Supplement are standard cell culture formulations containing a stabilized form of L-glutamine that prevents degradation and buildup of ammonia.
Other cell culture media
Gibco cell culture media are available in a variety of formulations to suit your research needs. Explore these additional options below.
IMDM media
Gibco Iscove’s Modified Dulbecco’s Medium (IMDM) is a highly enriched medium well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. Choose from a large selection of IMDM media compositions for a range of cell culture applications.
Media 199
Gibco Media 199 (M199 media) is useful across a wide range of species and applications but is particularly suitable for the culturing of non-transformed cells. M199 medium ingredients may include Earle’s salts and a bicarbonate/carbonic acid buffering system for the maintenance of pH in a CO2 incubator or Hank’s salts with salt solutions designed for atmospheric equilibrium.
Ham’s F-12
Gibco Ham’s F-12 Nutrient Mixture was formulated for single-cell plating of near-diploid Chinese Hamster Ovary (CHO) cells but has also been used for serum-free growth of CHO cultures and serum-supplemented growth of mammalian cells. Select from several high-quality and reliable formulations of Ham’s F-12 media, including glutamine-supplemented F-12 medium as well as GlutaMAX supplemented formulations, Kaighn's formulation, and powdered formats.
Ham’s F-10
Gibco Ham’s F-10 Nutrient Mixture is formulated for serum-free growth of Chinese Hamster Ovary (CHO) cells and the growth of various mammalian cell lines with serum supplementation. Rely on Gibco Ham's F-10 medium for superior quality, greater reliability, and improved consistency to give you better overall efficiency and more robust data.
Custom cell culture media
Customize Gibco media catalog products—add or remove components, create customized packaging options, select QC tests, and more. Media formulations available for customization include classical basal media, protein expression media, stem cell media, and cell and gene therapy (CGT) media.
Cell culture media for bioprocessing
Partnering with you to empower progress in bioprocessing around the world. Whether you are developing a vaccine or need help scaling up, our dedicated team will keep you at the forefront of the evolving bioprocessing industry.
Cell culture media by application
Regardless of whether you are performing cell culture in a research lab or production facility, need a special or defined formulation, or are growing cell lines, primary cells, or stem cells, Gibco products offer a reliable solution.
Organoid, spheroid, and 3D culture
Gibco media products have been used in a range of organoid, spheroid, and 3D culture studies, allowing for a more physiologically relevant understanding of complex biology.
Stem cell culture
An extensive selection of high-quality media products are available for pluripotent stem cells, neural stem cells, mesenchymal stem cells, hematopoietic stem cells, and stem cell therapy applications.
Neuronal cell culture
Gibco neuronal and neural cell media are optimized for the growth and support of primary neural cell and neuronal cultures and have been cited in applications involving primary neurons, glial cells, and disease models.
Primary cell culture
A wide range of Gibco media products are available for the support of many primary cell types, including fibroblasts, hepatocytes, and many more.
Cell Therapy Systems
Gibco Cell Therapy Systems (CTS) media products, designed for use in cell therapy applications, are manufactured to GMP standards and undergo extensive testing to ensure sterility and safety.
Protein expression media
Specific Gibco media formulations are available as components of our protein expression systems, supporting the generation of recombinant proteins for further use, including functional and therapeutic studies.
Media for cytogenetic research
A selection of specially formulated Gibco media are available for use in standard clinical cytogenetic protocols, with products optimized for analysis of amniotic fluid cells, chronic villus samples, bone marrow cells, or peripheral blood lymphocytes.
Serum-free media
Gibco serum-free media are formulated with specific combinations of nutrients and hormones that support the growth of certain cells in the absence of animal sera, allowing for increased selectivity of the media, easier purification, and more consistent performance.
Cell culture media best practices
Here are a few simple tips & tricks to help ensure your cell culture media are maintained for optimal performance.
If refrigerated, warm your cell culture media before use. Your cells would be cold at 2–8°C, and you would be too!
Keep your cell culture media protected from light. Light exposure degrades the essential vitamins in media that your cells need to grow.
Once you have supplemented the cell culture media with FBS, place the complete media into the refrigerator (2–8°C ) to maintain performance.
Use your supplemented media within 2–4 weeks to reduce the chances of contamination and the impact of pH drift.
Cell Culture Samples and Request a Quote Program
Simply complete the form and consult with a specialist to receive your sample(s) and/or quote.
GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Resources and support
Resources
Gibco Media Selection Tool
Find the right Gibco media for your application.  Use this interactive tool to select media based upon culture type, components, additives, and format. 
Cell Culture Select Tool
Search your cell line of interest to find product recommendations and resources to help enable a successful cell culture.
Gibco Cell Culture Basics
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Cell Culture Videos
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and the latest advancements in the field.
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.