rebranded sho-qubit-assays
Regardless of throughput, we've got a fluorescence-based quantification kit for your DNA and RNA samples. Utilizing simple mix-and-read protocols and incorporating fluorescence-based detection reagents, these assays are designed for accuracy and convenience.
Understand your sample’s purity and concentration
Understand your sample’s purity and concentration
The Thermo Scientific NanoDrop One UV-Vis Spectrophotometer and the Invitrogen Qubit fluorometers, when used together, provide the ability to obtain the most complete information about the concentration and quality of your DNA, RNA, or protein sample. Knowing the precise concentration and purity of your sample early helps you prevent costly troubleshooting and rework downstream.
Choose the platform that is right for you
Qubit assay kits for 1–20 samples
The Qubit assay kits for use with the Qubit 3.0 Fluorometer are designed for quantification of smaller numbers of samples (~1–20 samples, read in 0.2 mL tubes). Get rapid results with as little as 2 minutes' assay incubation and ~5 seconds' sample read time. Based on target-specific fluorescent dyes, Qubit assays offer accurate quantification data, even when samples contain common contaminants.
qubit-dsdna-hs-assay-s002330
Figure 1. Performance of the Qubit dsDNA HS Assay. The Qubit dsDNA HS Assay has a linear detection range of 0.2–100 ng and is selective for dsDNA, even in the presence of an equal mass of RNA.
Quant-iT assay kits
Quant-iT assay kits utilize advanced fluorophores that become fluorescent upon binding to DNA, RNA, or proteins; the fluorescence intensity of the resulting complex depends directly on the amount of the target molecule in the sample.
Quant-iT assay kits for 20–2,000 samples
The Quant-iT Kits, 1,000 assay size, provide the ultimate in convenience for DNA, RNA, or protein quantitation in microplates. These are the best choice for quantitating 20–2,000 samples at a time.
Quantitation using Quant-iT Assay Kits
Figure 2. Quantitation using Quant-iT Assay Kits. (Left) The Quant-iT dsDNA Assay Kit, High Sensitivity has a linear detection range of 0.2–100 ng and is selective for dsDNA, even in the presence of an equal mass of RNA. The x-axis gives the mass of nucleic acid when DNA or RNA is assayed alone; in the 1:1 mixture, the total mass of nucleic acid is double the amount shown. The inset shows the sensitivity of the assay for DNA. (Center) The Quant-iT RNA Assay Kit has a linear detection range of 5–100 ng and is selective for RNA, even in the presence of an equal mass of DNA. The x-axis gives the mass of nucleic acid when DNA or RNA is assayed alone; in the 1:1 mixture, the total mass of nucleic acid is double what is stated. (Right) The Quant-iT Protein Assay Kit has a detection range of 0.25–5 μg. This graph shows the signal generated from nine different proteins over the stated detection range. The protein-to-protein variability at the 3 μg point for all nine proteins assayed is 12%.
Quant-iT bulk reagents and kits for >20,000 samples
These Quant-iT products provide the maximum amount of dye in packaging that is amenable to highly automated quantitation environments. These are the best choice for quantitating more than 2,000 samples at a time.
These products are available in either concentrated Quant-iT dyes or Quant-iT kits with buffer and a standard included.
Nucleic acid quantitation using Quant-iT Reagents
Figure 3. Nucleic acid quantitation using Quant-iT Reagents. (Left) The fluorescence signal from binding of the Quant-iT PicoGreen dsDNA Reagent to dsDNA is linear over at least four orders of magnitude; the inset shows the results obtained using 0–750 pg/mL calf thymus DNA. (Center) The Quant-iT RiboGreen RNA Assay provides linear quantitation from 20 ng/mL to 1 μg/mL RNA using a 400-fold dilution of the dye; the inset shows the results for 1.0 ng/mL to 50 ng/mL RNA obtained using a 4,000-fold dilution of the dye. (Right) Quantitation of a synthetic 24-mer using the Quant-iT OliGreen ssDNA Reagent is linear over at least four orders of magnitude; the inset shows the results obtained using 0–2.0 ng/mL of oligo.
Qubit assay kits for 1–20 samples
Catalog # Name Size Price (USD) Qty
Q32850 Qubit™ dsDNA BR Assay Kit, 100 assays Each
147.00
Q32853 Qubit™ dsDNA BR Assay Kit, 500 assays Each
435.00
Q32851 Qubit™ dsDNA HS Assay Kit, 100 assays Each
147.00
Q32854 Qubit™ dsDNA HS Assay Kit, 500 assays Each
437.00
Q10212 Qubit™ ssDNA Assay Kit Each
146.00
Q32852 Qubit™ RNA HS Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q32855 Qubit™ RNA HS Assay Kit, 500 assays Each
406.65
Online Exclusive
430.00
Save 23.35 (5%)
Q10210 Qubit™ RNA BR Assay Kit, 100 assays Each
139.65
Online Exclusive
145.00
Save 5.35 (4%)
Q10211 Qubit™ RNA BR Assay Kit, 500 assays Each
408.65
Online Exclusive
431.00
Save 22.35 (5%)
Q32880 Qubit™ microRNA Assay Kit, 100 assays Each
139.65
Online Exclusive
146.00
Save 6.35 (4%)
Q32881 Qubit™ microRNA Assay Kit, 500 assays Each
421.65
Online Exclusive
444.00
Save 22.35 (5%)
Q33211 Qubit™ Protein Assay Kit, 100 Assays Each
149.65
Online Exclusive
158.00
Save 8.35 (5%)
Q33212 Qubit™ Protein Assay Kit, 500 Assays Each
447.65
Online Exclusive
476.00
Save 28.35 (6%)
Quant-iT™ assay kits for 20–2,000 samples
Catalog # Name Size Price (USD) Qty
Q33120 Quant-iT™ dsDNA Assay Kit, high sensitivity, 1 kit Each
596.65
Online Exclusive
650.00
Save 53.35 (8%)
Q33130 Quant-iT™ dsDNA Assay Kit, broad range, 1 kit Each
607.65
Online Exclusive
658.00
Save 50.35 (8%)
Q33140 Quant-iT™ RNA Assay Kit, 1 kit Each
580.65
Online Exclusive
624.00
Save 43.35 (7%)
Q10213 Quant-iT™ RNA Assay Kit, broad range, 1,000 assays Each
592.65
Online Exclusive
624.00
Save 31.35 (5%)
Q32882 Quant-iT™ microRNA Assay Kit Each
735.65
Online Exclusive
758.00
Save 22.35 (3%)
Q33210 Quant-iT™ Protein Assay Kit Each
609.65
Online Exclusive
654.00
Save 44.35 (7%)
Quant-iT bulk reagents and kits for >20,000 samples
Catalog # Name Size Price (USD) Qty
P7581 Quant-iT™ PicoGreen™ dsDNA Reagent, 1 mL Each
592.65
Online Exclusive
620.00
Save 27.35 (4%)
P7589 Quant-iT™ PicoGreen™ dsDNA Assay Kit, 1 mL kit Each
667.65
Online Exclusive
700.00
Save 32.35 (5%)
R11491 Quant-iT™ RiboGreen™ RNA Reagent, 1 mL Each
711.65
Online Exclusive
754.00
Save 42.35 (6%)
R11490 Quant-iT™ RiboGreen™ RNA Assay Kit, 1 kit Each
758.65
Online Exclusive
800.00
Save 41.35 (5%)
O7582 Quant-iT™ OliGreen™ ssDNA Reagent, 1 mL Each
726.65
Online Exclusive
778.00
Save 51.35 (7%)
O11492 Quant-iT™ OliGreen™ ssDNA Assay Kit, 1 kit Each
771.65
Online Exclusive
826.00
Save 54.35 (7%)
P11495 Quant-iT™ PicoGreen™ dsDNA Reagent, 10 x 100 μL Each
713.65
Online Exclusive
748.00
Save 34.35 (5%)
P11496 Quant-iT™ PicoGreen™ dsDNA Assay Kit, 10 x 100 μL kit Each
738.65
Online Exclusive
774.00
Save 35.35 (5%)
* Invitrogen Quant-iT assays and reagents can also be used with the Invitrogen Qubit Fluorometer.