Immunoprecipitation (IP) is a key technique in cellular and molecular biology, employed to isolate or purify specific antigens (proteins of interest) using solid phases like agarose or Sepharose™ resin, or magnetic particles. The IP method was initially performed on small aliquots of agarose resin and is used when scale-up is anticipated. Magnetic beads are versatile, enabling the targeting of single proteins or peptides (IP), studying protein interactions through protein complexes (Co-IP), identify regions of the genome with DNA-binding or RNA-binding proteins (ChIP or RIP), and exploring the functional dynamics of protein-nucleic acid interactions.
Advantages of magnetic beads for immunoprecipitation
Magnetic beads for IP have become more common, especially compared to the traditional method of using agarose slurries or resin for protein isolation. This is because magnetic immunoprecipitation beads provide several advantages over the traditional method, such as:
  • Low background—Little to no non-specific binding and no preclearing required for high-purity protein yields
  • Highly reproducible—Uniform composition of the magnetic beads helps ensure the most consistent results
  • Highly sensitiveMost-cited method for sensitive applications, such as ChIP and IP for low- abundance proteins
  • Fast and easy—<40 min protocol with no centrifugation or preclearing steps
  • Antibody savings—All binding occurs on the smooth outer surface of the beads, which conserves precious antibodies compared to porous agarose beads; using beads for IP is a cost-efficient solution per sample
  • Automation compatible—Immunoprecipitation beads protocol can be automated on the Thermo Fisher Scientific KingFisher platform
View common IP myths on agarose resins vs magnetic beads
Learn about co-immunoprecipitation
Immunoprecipitation beads selection guides
Antibody-binding magnetic beads for IP
 Protein A, G, A/GSecondary antibodies
(anti-mouse, anti-rabbit)
Recommended productsBeads only:Kits:Beads only:
Binding propertiesNon-covalent antibody binding
Antibody co-eluted off the beadsYes*
No (Pierce Crosslink Magnetic IP/Co-IP Kit)
Yes*
Type of ligandDifferent ligands bind different species and antibody subclasses with different specificity
  • Anti-mouse binds mouse IgG1, IgG2a, IgG2b
  • Anti-rabbit binds all rabbit IgGs
Non-specific bindingLow
* Crosslinking can be performed to avoid co-elution of the antibody, but this can decrease the yield of the target antigen.
† Learn which antibody-binding proteins are best for your IP antibody.
Learn more about how mass spectrometry and western workflows are enhanced by high-performance protein enrichment using Dynabeads and DynaGreen magnetic beads in our magnetic bead product brochure
Dynabeads magnetic beads compared to Sepharose- and agarose-based solutions
Figure 1. Shorter protocol time and better target protein yields with Dynabeads magnetic beads. Equal amounts of sample material with respect to Ab content and cell lysate volume were used for all IP protocols according to the manufacturer’s protocol. For the Dynabeads-based method, all the antibodies on the bead surface are accessible for optimal, highly reproducible antigen binding.
Dynabeads magnetic beads compared to competitor magnetic beads
Figure 2. Electron microscopy of Dynabeads magnetic beads compared to competitor magnetic beads. Dynabeads and DynaGreen magnetic beads have a defined surface to carry out the necessary binding, with no inner surface to trap any unwanted proteins. Dynabeads products (A) are the most uniform, monodisperse superparamagnetic beads, manufactured with highly controlled product qualities to help ensure the highest degree of purity and reproducibility, while the DynaGreen products (B) offer high purity and higher binding capacity vs. the Dynabeads products, but also offer a more sustainability-conscious version of Dynabeads products. Magnetic particles from alternative suppliers (C–E) have variable shapes and sizes that trap impurities, resulting in lower purity and reproducibility without proper functionalization and antibody coupling.
Dynabeads magnetic beads compared to competitor magnetic beads for IP
Figure 3. Dynabeads magnetic beads have exceptional immunoprecipitation performance with the combination of highest yield and lowest non-specific binding. (A) Silver staining of the protein membrane shows that Dynabeads magnetic beads have the most enriched binding at the antibody main band and very low non-specific binding with low signal/noise. (B) Western blot analysis of CD81 pulldown in Jurkat cells show that Dynabeads Protein G beads have the highest yield with a short ten-minute protein incubation.
Read tips on immunoprecipitation
Fusion tag-binding magnetic beads for IP
Tagc-MycFLAGHAHistidineGST
Description
  • Consists of a peptide (EQKLISEEDL) derived from the human c-myc oncogene (p62 c-myc)
  • Immobilized anti-c-Myc antibodies are used to immunoprecipitate c-Myc-tagged recombinant proteins
  • Consists of a peptide (DYKDDDDK) which is recognized by an immobilized high-affinity rat monoclonal antibody (clone L5)
  • Primarily used for the isolation of protein complexes with multiple subunits because the mild purification process tends not to disrupt these interactions
  • Consists of a peptide (YPYDVPDYA) derived from the human influenza hemagglutinin (HA) protein
  • Immobilized anti-HA antibodies are used to immunoprecipitate HA-tagged recombinant proteins
  • Consists of a string of six to nine histidine residues
  • Primarily used for purification via immobilized metal affinity chromatography (IMAC)
  • Consists of glutathione S-transferase (GST), a complete 211 amino acid protein (26 kDa)
  • Primarily used for purification via glutathione agarose resin
Recommended products
His and GST are not true epitope tags, because they are not usually purified or detected via specific antibodies. By contrast, HA and c-Myc tags are true epitope tags, because their only means of purification or detection is via specific antibodies. Epitope tags are seldom used for bulk purification but are most often used for IP or co-IP. Nevertheless, all four of these tag systems can be used for either purification or pull-down applications.
View fusion tag-binding bead options for IP
Biotin-binding magnetic beads for IP
If you have a biotinylated antibody (or ligand) that recognizes your protein, use of streptavidin beads is recommended
Main advantages of magnetic beads for biotin-binding:
  • If you have a sample rich in soluble IgGs
  • If you have a recombinant antibody lacking Fc regions
  • If you need a bead compatible with mass spectrometry
  •  
    Recommended productsDynabeads M-280 StreptavidinDynabeads MyOne Streptavidin T1*Pierce Streptavidin Magnetic Beads
    Description
    • Hydrophobic bead surface
    • Based on tosylactivated beads
    • Size distribution: CV <3%
    • BSA as blocking protein
    • Isoelectric point: pH 5.0
    • Low charge (–10 mV at pH 7)
    • Iron content (Ferrites): 12% (17%)
    • Hydrophobic bead surface
    • Based on tosylactivated beads
    • Size distribution: CV <3%
    • BSA as blocking protein
    • Isoelectric point: pH 5.0
    • Low charge (–10 mV at pH 7)
    • Iron content (Ferrites): 26% (37%)
    • Hydrophilic bead surface
    • Size distribution: 0.7–1.7 mm
    • Covalently pre-blocked (non-protein) to minimize nonspecific binding
    • pH range: 4–11
    • Iron content (Fe3O4): 60% (double layer of magnetite)
    • Particle density: ~2.0 g/cm3
    Capacity
    • Free biotin: 490–750 pmol/mg beads
    • Biotinylated Ig: 5–10 µg/mg beads
    • Free biotin: 950–1,500 pmol/mg beads
    • Biotinylated Ig: 20 µg/mg beads
    • Biotinylated fluorescein: 3500 pmol/mg
    • Biotinylated IgG: 55 mg/mg
    * Low sedimentation rate and improved reaction kinetics compared to M-280 beads.
    View streptavidin bead options for IP
    Surface-activated magnetic beads for IP
    If your target protein or ligand needs to be covalently coupled, use surface-activated beads
    Different ligands (antibodies, proteins, etc.) require different bead surface properties and immobilization chemistries. Choose surface-activated beads to control factors such as:
    • Hydrophobicity: choose hydrophilic of hydrophobic beads
    • Bead size: 1 µm, 2.8 µm, and 4.5 µm beads available
    • Surface chemistry: epoxy, amine, or tosylactivated
    View surface-activated bead options for IP
    Searching for high-performing, sustainable magnetic beads? Introducing DynaGreen Magnetic Beads.
    DynaGreen Magnetic Beads provide high-quality, reproducible results and are also environmentally friendly and microplastic-free. How do these beads contribute to a more sustainable science? All DynaGreen Magnetic Beads have the following features:
    • Non-microplastic bead core
    • Energy-efficient manufacturing
    • Reduced water consumption
    • Use of non-hazardous chemicals
    • Recyclable packaging
    • Paperless documents
    Learn more about DynaGreen Magnetic Beads
    Detect and analyze proteins in as little as 40 minutes
    With automation protocols available for KingFisher instruments, you can be ready to detect and analyze proteins in as little as 40 minutes with minimal hands-on time.
    Learn more about KingFisher automation
    What is immunoprecipitation?
    Immunoprecipitation (IP) is a cellular and molecular biology method used in laboratories to isolate or purify a specific antigen (or protein of interest) via a solid phase (agarose or Sepharose™ resin or magnetic particles). The IP method was first performed on small aliquots of agarose resin in microcentrifuge tubes. The IP method has evolved to use superparamagnetic particles to specifically target proteins of interest.
    View common IP myths and tips
    How does immunoprecipitation work?
    The Dynabeads magnetic beads IP procedure is simple, fast, requires no preclearing step, and results in high-target protein purity, yield, and consistent results. Both DynaGreen magnetic beads and Pierce magnetic beads can perform the same steps with slightly longer protocol times to complete. DynaGreen protocol finishes in <80 min. Pierce protocol finishes in <120 min.
    Step 1: Binding of the antibody to the beads
    Dynabeads, DynaGreen, and Pierce magnetic beads come pre-coated with protein A, protein G, anti-mouse IgG, or anti-rabbit IgG antibodies. Their rapid kinetics allow them to bind to the added antibody in about ten minutes (Figure 4). Biotinylated antibodies can also be used in combination with streptavidin-coupled Dynabeads and Pierce magnetic beads. After antibody incubation a wash step is performed to remove unbound antibodies.
    Step 2: Add sample to the beads
    The protein-containing sample is added to the beads-antibody mix and incubated for ten minutes to allow target protein binding. If the protein is of low abundance or low affinity, the incubation time can be increased to one hour or overnight. For most cases, a ten-minute incubation is sufficient.
    Step 3: Wash the protein-bound beads
    To help ensure high purity of the bound target protein, the beads should be washed 2 to 4 times with buffer on the magnet to remove all unbound proteins. This step in the isolation process helps ensure a high signal-to-noise ratio.
    Step 4: Elute the protein
    The protein can be eluted off the beads by using either a mild elution procedure or a denaturing elution procedure (Figure 5).
    Figure 4. Immunoprecipitation protocol with magnetic beads.
    Chart demonstrating denaturing elution is best used during SDS-PAGE; mild elution is best used for protein characterization, immunization, enzyme studies, amino acid sequence determination, and crystallization; and no elution is needed when performing protein-protein interactions studies, enzyme studies, bioassays, and immunoassays
    Figure 5. Types of elution buffers for immunoprecipitation with magnetic beads. Depending on your target protein and how they bind to the magnetic beads, there are three main types of elution buffers to unbind the target protein. Denaturing elution buffers are used for SDS-PAGE. Mild elution buffers are used for protein characterization, immunization, enzyme studies, amino acid sequence determination, and crystallization. Protein-protein interactions, enzyme studies, bioassays, and immunoassays may be performed without eluting your target protein from the magnetic beads.
    Videos on immunoprecipitation
    Covalent Coupling of Antibodies to Magnetic Beads - Fast and Efficient
    The benefits of the Dynabeads Antibody CoupliKit include: • Use any primary antibody • Get ultra-low non-specific binding • No need for cross-linking since Antibody is irreversibly bound to the beads and will not be eluted off with target protein
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
          1. Now Playing
            Up NextCovalent Coupling of Antibodies to Magnetic Beads - Fast and Efficient
            The benefits of the Dynabeads Antibody CoupliKit include: • Use any primary antibody • Get ultra-low non-specific binding • No need for cross-linking since Antibody is irreversibly bound to the beads and will not be eluted off with target protein
          2. Now Playing
            Up NextIP Myth 4 Dynabeads are expensive
            IP Myth 4 Dynabeads are expensive
          3. Now Playing
            Up NextIP Myth 3 Capacity is crucial
            IP Myth 3 Capacity is crucial
          4. Now Playing
            Up NextIP Myth 2 Pre-clearing is necessary
            IP Myth 2 Pre-clearing is necessary
          5. Now Playing
            Up NextIP Myth 1 Background can't be avoided
            IP Myth 1 Background can't be avoided
          6. Now Playing
            Up NextHow To Use Dynabeads For Immunoprecipitation
            How to use Dynabeads for immunoprecipitation
          7. Now Playing
            Up NextAutomated Immunoprecipitation in 40 min using Dynabeads and KingFisher Flex
            For IP, KingFisher instruments with Dynabeads offer the best balance of high yield and reproducibility with low nonspecific binding and cost, which is one reason why they have become the gold standard for immunoprecipitation using magnetic beads.
          8. Now Playing
            Up NextImmunoprecipitation Publication Trends - The Reasons for the Shift
            Immunoprecipitation Publication Trends - The Reasons for the Shift
        Immunoprecipitation with Dynabeads magnetic beads FAQs
        Here are some frequently asked questions regarding the use of Dynabeads magnetic beads for immunoprecipitation.
        A: Dynabeads Protein A and Dynabeads Protein G are 2.8-micron beads covalently coupled with Protein A and recombinant Protein G, respectively. Protein A and Protein G differ in their binding strength to immunoglobulins from different species and subclasses.
        View the Dynabeads product selection guide for detailed information on Ig species and subclass specificity to Dynabeads
        A: 1. Verify binding/specificity of your antibody to your antigen (e.g., by ELISA).
        2. Check the binding of your antibodies to the beads. If the antibodies are not captured and bound to the beads, the immunoprecipitation experiment will not work.
        3. If you have used the indirect method of immunoprecipitation, try the direct method. Conversely, if you have used the direct method, try the indirect method.
        4. Check the amount of beads and sample volume. With reference to the capacity of different beads proposed in the package inserts, increase the amount of beads or the concentration of your antibody during coupling.
        5. Increase the incubation time.
        6. Try another antibody.
        A: Yes, you can use the larger beads. Note: Smaller beads provide a larger surface area and therefore give higher yields of protein compared to the larger 4.5-micron beads.
        Immunoprecipitation publications using Dynabeads magnetic beads
        View publications on cell isolation using Dynabeads products
        View publications on Dynabeads Streptavidin magnetic beads
        Additional publications can be found in our Citations and References database. The database can be searched using a product catalog number or keyword. For example, searching with the term "10001D" will return a list of journal articles that cite the use of Dynabeads Protein A for Immunoprecipitation (Cat. No. 10001D) and searching with the term "dynabeads" will return a list of journal articles that cite the use of any Dynabeads product. Note that the Citations and References database on thermofisher.com contains only the small subset of scientific journal articles that cite the use of our products. Many more can be found via targeted searches of the PubMed™ database and similar databases.
        Ordering information
        Catalog # Name Size Price (USD) Qty
        10001D Dynabeads™ Protein A for Immunoprecipitation, 1 mL Each
        233.65
        Online Exclusive
        250.00
        Save 16.35 (7%)
        10002D Dynabeads™ Protein A for Immunoprecipitation, 5 mL Each
        876.65
        Online Exclusive
        938.00
        Save 61.35 (7%)
        10008D Dynabeads™ Protein A for Immunoprecipitation, 50 mL Each
        5,960.65
        Online Exclusive
        6,410.00
        Save 449.35 (7%)
        10003D Dynabeads™ Protein G for Immunoprecipitation, 1 mL Each
        238.65
        Online Exclusive
        250.00
        Save 11.35 (5%)
        10004D Dynabeads™ Protein G for Immunoprecipitation, 5 mL Each
        877.65
        Online Exclusive
        938.00
        Save 60.35 (6%)
        10009D Dynabeads™ Protein G for Immunoprecipitation, 50 mL Each
        5,955.65
        Online Exclusive
        6,410.00
        Save 454.35 (7%)
        78609 Pierce™ Protein A/G Magnetic Agarose Beads, 4 mL Each
        481.65
        Online Exclusive
        536.00
        Save 54.35 (10%)
        78610 Pierce™ Protein A/G Magnetic Agarose Beads, 20 mL Each
        1,656.65
        Online Exclusive
        1,846.00
        Save 189.35 (10%)
        88802 Pierce™ Protein A/G Magnetic Beads, 1 mL Each
        228.65
        Online Exclusive
        260.00
        Save 31.35 (12%)
        88803 Pierce™ Protein A/G Magnetic Beads, 5 mL Each
        861.65
        Online Exclusive
        976.00
        Save 114.35 (12%)
        10006D Dynabeads™ Protein A Immunoprecipitation Kit Each
        528.00
        10007D Dynabeads™ Protein G Immunoprecipitation Kit Each
        528.00
        88804 Pierce™ Classic Magnetic IP/Co-IP Kit Each
        511.65
        Online Exclusive
        572.00
        Save 60.35 (11%)
        88805 Pierce™ Crosslink Magnetic IP/Co-IP Kit Each
        618.65
        Online Exclusive
        676.00
        Save 57.35 (8%)
        90409 Pierce™ MS-Compatible Magnetic IP Kit, Protein A/G, 40 reactions kit Each
        558.65
        Online Exclusive
        630.00
        Save 71.35 (11%)
        11201D Dynabeads™ M-280 Sheep Anti-Mouse IgG, 2 mL Each
        540.00
        11202D Dynabeads™ M-280 Sheep Anti-Mouse IgG, 10 mL Each
        1,680.00
        11203D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 2 mL Each
        540.00
        11204D Dynabeads™ M-280 Sheep Anti-Rabbit IgG, 10 mL Each
        1,680.00
        80101G DynaGreen™ Protein A Magnetic Beads, 0.5 mL, 20 rxns Each
        143.65
        Online Exclusive
        148.00
        Save 4.35 (3%)
        80102G DynaGreen™ Protein A Magnetic Beads, 3 mL, 120 rxns Each
        672.65
        Online Exclusive
        706.00
        Save 33.35 (5%)
        80103G DynaGreen™ Protein A Magnetic Beads, 25 mL, 1000 rxns Each
        4,285.65
        Online Exclusive
        4,415.00
        Save 129.35 (3%)
        80104G DynaGreen™ Protein A/G Magnetic Beads, 0.5 mL, 20 rxns Each
        148.00
        80105G DynaGreen™ Protein A/G Magnetic Beads, 3 mL, 120 rxns Each
        706.00
        80106G DynaGreen™ Protein A/G Magnetic Beads, 25 mL, 1000 rxns Each
        4,415.00
        80108G DynaGreen™ CaptureSelect™ Anti-IgG-Fc (Multi-Species) Magnetic Beads, 3 mL, 120 rxns Each
        667.65
        Online Exclusive
        706.00
        Save 38.35 (5%)
        80107G DynaGreen™ CaptureSelect™ Anti-IgG-Fc (Multi-Species) Magnetic Beads, 0.5 mL, 20 rxns Each
        140.65
        Online Exclusive
        148.00
        Save 7.35 (5%)
        80109G DynaGreen™ CaptureSelect™ Anti-IgG-Fc (Multi-Species) Magnetic Beads, 25 mL, 1000 rxns Each
        4,195.65
        Online Exclusive
        4,415.00
        Save 219.35 (5%)
        Catalog # Name Size Price (USD) Qty
        10103D Dynabeads™ His-Tag Isolation and Pulldown, 2 mL Each
        272.65
        Online Exclusive
        285.00
        Save 12.35 (4%)
        10104D Dynabeads™ His-Tag Isolation and Pulldown, 10 mL Each
        1,132.65
        Online Exclusive
        1,186.00
        Save 53.35 (4%)
        78605 Pierce™ Ni-NTA Magnetic Agarose Beads, 4 mL Each
        283.65
        Online Exclusive
        319.00
        Save 35.35 (11%)
        78606 Pierce™ Ni-NTA Magnetic Agarose Beads, 20 mL Each
        1,052.65
        Online Exclusive
        1,170.00
        Save 117.35 (10%)
        88831 HisPur™ Ni-NTA Magnetic Beads, 2 mL Each
        285.65
        Online Exclusive
        313.00
        Save 27.35 (9%)
        88832 HisPur™ Ni-NTA Magnetic Beads, 10 mL Each
        1,014.65
        Online Exclusive
        1,162.00
        Save 147.35 (13%)
        78601 Pierce™ Glutathione Magnetic Agarose Beads, 4 mL Each
        206.65
        Online Exclusive
        229.00
        Save 22.35 (10%)
        78602 Pierce™ Glutathione Magnetic Agarose Beads, 20 mL Each
        686.65
        Online Exclusive
        768.00
        Save 81.35 (11%)
        88836 Pierce™ Anti-HA Magnetic Beads, 1 mL Each
        329.65
        Online Exclusive
        377.00
        Save 47.35 (13%)
        88837 Pierce™ Anti-HA Magnetic Beads, 5 mL Each
        1,056.65
        Online Exclusive
        1,246.00
        Save 189.35 (15%)
        88838 Pierce™ HA-Tag Magnetic IP/Co-IP Kit Each
        583.65
        Online Exclusive
        664.00
        Save 80.35 (12%)
        88842 Pierce™ Anti-c-Myc Magnetic Beads Each
        435.65
        Online Exclusive
        485.00
        Save 49.35 (10%)
        88844 Pierce™ c-Myc-Tag Magnetic IP/Co-IP Kit Each
        776.00
        A36797 Pierce™ Anti-DYKDDDDK Magnetic Agarose, 4 mL Each
        460.65
        Online Exclusive
        514.00
        Save 53.35 (10%)
        A36798 Pierce™ Anti-DYKDDDDK Magnetic Agarose, 20 mL Each
        1,476.65
        Online Exclusive
        1,676.00
        Save 199.35 (12%)
        Catalog # Name Size Price (USD) Qty
        11205D Dynabeads™ M-280 Streptavidin, 2 mL Each
        663.65
        Online Exclusive
        724.00
        Save 60.35 (8%)
        11206D Dynabeads™ M-280 Streptavidin, 10 mL Each
        2,112.65
        Online Exclusive
        2,305.00
        Save 192.35 (8%)
        60210 Dynabeads™ M-280 Streptavidin, 100 mL Each
        11,668.65
        Online Exclusive
        12,730.00
        Save 1,061.35 (8%)
        65601 Dynabeads™ MyOne™ Streptavidin T1, 2 mL Each
        647.65
        Online Exclusive
        702.00
        Save 54.35 (8%)
        65602 Dynabeads™ MyOne™ Streptavidin T1, 10 mL Each
        2,145.65
        Online Exclusive
        2,325.00
        Save 179.35 (8%)
        65604D Dynabeads™ MyOne™ Streptavidin T1, 50 mL Each
        8,783.65
        Online Exclusive
        9,520.00
        Save 736.35 (8%)
        88817 Pierce™ Streptavidin Magnetic Beads, 5 mL Each
        1,170.65
        Online Exclusive
        1,342.00
        Save 171.35 (13%)
        88816 Pierce™ Streptavidin Magnetic Beads, 1 mL Each
        358.65
        Online Exclusive
        405.00
        Save 46.35 (11%)
        Catalog # Name Size Price (USD) Qty
        14301 Dynabeads™ M-270 Epoxy, 60 mg Each
        530.65
        Online Exclusive
        578.00
        Save 47.35 (8%)
        14302D Dynabeads™ M-270 Epoxy, 300 mg Each
        1,799.65
        Online Exclusive
        1,962.00
        Save 162.35 (8%)
        14011 Dynabeads™ M-450 Epoxy Each
        960.00
        14311D Dynabeads™ Antibody Coupling Kit Each
        635.65
        Online Exclusive
        666.00
        Save 30.35 (5%)
        14321D Dynabeads™ Co-Immunoprecipitation Kit Each
        970.65
        Online Exclusive
        1,026.00
        Save 55.35 (5%)
        88826 Pierce™ NHS-Activated Magnetic Beads, 1 mL Each
        312.65
        Online Exclusive
        348.00
        Save 35.35 (10%)
        88827 Pierce™ NHS-Activated Magnetic Beads, 5 mL Each
        1,058.65
        Online Exclusive
        1,152.00
        Save 93.35 (8%)
        88828 Pierce™ Direct Magnetic IP/Co-IP Kit Each
        578.65
        Online Exclusive
        632.00
        Save 53.35 (8%)
        65501 Dynabeads™ MyOne™ Tosylactivated, 2 mL Each
        1,255.65
        Online Exclusive
        1,344.00
        Save 88.35 (7%)
        65502 Dynabeads™ MyOne™ Tosylactivated, 10 mL Each
        4,072.65
        Online Exclusive
        4,360.00
        Save 287.35 (7%)
        14203 Dynabeads™ M-280 Tosylactivated, 2 mL Each
        493.65
        Online Exclusive
        554.00
        Save 60.35 (11%)
        14204 Dynabeads™ M-280 Tosylactivated, 10 mL Each
        1,459.65
        Online Exclusive
        1,660.00
        Save 200.35 (12%)
        14013 Dynabeads™ M-450 Tosylactivated Each
        1,082.00
        Selection guide for Dynabeads products
        Dynabeads products are available for a multitude of molecular and cellular applications. Don’t know where to start? Use our easy selection guide that considers sample type, target molecule, and downstream application to provide tailored recommendations for Dynabeads products.
        Dynabeads product selection guide
        Hero banner dark gradient overlay
        ACT is a trademark of My Green Lab.
        Magne is a trademark of Promega Corporation.
        Sepharose is a trademark of Cytiva Bioprocess R&D AB.
        SureBeads is a trademark of Bio-Rad Laboratories, Inc.
        Sera-Mag is a trademark of Hyclone Laboratories, LLC.
        GenScript is a trademark of Nanjing GenScript Biotech Co.
        PureProteome is a trademark of Millipore Corporation.
        Stylesheet for Classic Wide Template adjustments
        For Research Use Only. Not for use in diagnostic procedures.