Search Thermo Fisher Scientific
Save these protocols on how to mix media and reagents from powders or concentrates.
In addition to the materials listed below, you will also need:
While powdered media takes additional time to prepare, cost savings and extended shelf life make it a good choice for many labs. Media concentrates are also a great alternative to pre-diluted formulas because they are easier to store and handle compared to powdered media, while offering convenience and flexibility for precise dilution to desired concentrations.
To make cell culture media from powder, refer to the generic protocol below. Precise quantities and volumes of the pH adjustors for your chosen cell culture media can be found on Table 1.
The following protocol is a generic powdered cell culture media preparation protocol. To prepare DMEM from powder—or any additional media formulations, tailor this protocol with the values provided in Table 1.
Media | Cat. No. | Sodium bicarbonate(NaHCO3) | Recommended working pH | |
---|---|---|---|---|
g/L | mL/L 7.5% soln. | |||
Grace's Insect Medium, unsupplemented, powder | 0.35 | 4.7 | 5.9–6.2 | |
MEM, powder | 61100053 61100061 61100087 61100103 | 2.2 | 29.3 | 7.0–7.4 |
MEM, powder, autoclavable, no glutamine | Not recommended | 30.0 | 7.2–7.4 | |
Leibovitz's L-15, powder | 41300021 41300039 41300070 | Do not add | 7.6 | |
Iscove’s Modified Dulbecco’s Medium (IMDM) | 12200036 12200069 | 3.024 | Not recommended | Do not adjust |
MEM, glutamine, HEPES, powder | 0.85 | 11.35 | 7.0–7.4 | |
MEM, Hanks' Balanced Salts, powder | 0.35 | 4.7 | 7.0–7.4 | |
MEM α, nucleosides, powder | 11900016 11900024 11900073 | 2.2 | 29.3 | 7.0–7.4 |
MEM α, no nucleosides, powder | 12000014 12000022 12000063 | 2.2 | 29.3 | 7.0–7.4 |
MEM, NEAA, powder | 41500018 41500034 41500067 41500083 | 2.2 | 29.3 | 7.0–7.4 |
TC 100 Medium, powder
| 0.35 | 4.7 | 6.1–6.3 | |
DMEM, powder, high glucose | 52100021 52100039 52100047 | 3.7 | 49.3 | 7.0–7.4 |
DMEM/F-12, powder, HEPES | 12400024 42400010 42400028 | 1.2 | 16.0 | 7.0–7.4 |
DMEM/F-12, powder | 12500062 12500096 | 2.438 | 32.5 | 7.0–7.4 |
DMEM, powder, high glucose, pyruvate | 12800017 12800082 | 3.7 | 49.3 | 7.0–7.4 |
DMEM, powder, low glucose, pyruvate | 31600034 31600083 31600091 | 3.7 | 49.3 | 7.0–7.4 |
Ham's F-12 Nutrient Mix, powder | 21700018 21700026 21700075 | 1.18 | 15.7 | 7.5–7.9 |
Glasgow's MEM, powder | 2.75 | 36.6 | 6.7–7.1 | |
Opti-MEM Reduced Serum Medium, powder | 22600134 22600050 | 2.4 | 32.0 | 7.1–7.4 |
RPMI 1640 Medium, powder, HEPES | 13018015 13018031 | 2.0 | 26.7 | 7.0–7.4 |
RPMI 1640 Medium, powder | 31800022 31800089 31800105 51800019 51800035 51800043 | 2.0 | 26.7 | 7.0–7.4
|
Medium 199, Hank's, powder
| 10012011 10012037 | .35 | 4.7 | 7.0–7.4 |
Medium 199, Earle's Salts, powder | 31100019 31100027 31100035 | 2.2 | 29.3 | 7.1–7.4 |
Dipotassium hydrogen phosphate, 98.0-100.5% (dried basis), USP, Multi-Compendial, Endotoxin-free, GMP, J.T.Baker™ | 3250-01 3250-05 3250-06 3250-07 | 2.43 | 32.5 | 7.1–7.4 |
PFHM-II Protein-Free Hybridoma Medium (powder) | 26.7 | 6.8-7.2 |
The following is a general cell culture media preparation protocol for 1X solutions from common 10X liquid media concentrates. Please note, because we adjust the pH of all 10X media and 10X balanced salt solution concentrates for solubility, you may need to adjust the pH after the dilution and to add sodium bicarbonate, as appropriate.
Media | Cat. No. | pH of 10X solution | Sodium bicarbonate(NaHCO3) | pH after 1:10 dilution & NaHCO3 addition* | pH of 1X solution* | |
---|---|---|---|---|---|---|
g/L | mL/L 7.5% soln. | |||||
MEM (10X), no glutamine | 11430030 | 4.8–5.3 | 2.2 | 29.3 | 7.6–7.9 | 7.0–7.4 |
Medium 199 (10X) | 11825015 | 2.4–2.7 | 2.2 | 29.3 | 7.6–7.9 | 7.1–7.4 |
Medium 199, Earle's Salts (10X) | 21180021 | 2.0–2.8 | 2.2 | 29.3 | 7.6–7.9 | 7.1–7.4 |
MEM (10X), no glutamine | 21430020 21430079 | 4.8–5.3 | 2.2 | 29.3 | 7.6–7.9 | 7.0–7.4 |
* pH range varies depending on the pH of water used in dilution.
Powdered reagents and liquid concentrates, such as DPBS (Dulbecco’s Phosphate Buffered Saline), are a good option for labs that want to save money and storage space because they are typically less expensive than traditional liquid reagents and take up less room. Powdered reagents also have a longer shelf life, and allow personnel to prepare only the amount needed, minimizing the risk of reagent waste due to expiration.
The following protocol prepares a 1x concentration of powdered Dulbecco’s Phosphate Buffered Saline with calcium and magnesium.
To prepare an acceptable, final 1X solution from 10X concentrates of cell culture reagents, perform the following procedure under aseptic conditions. Use distilled water and 7.5% sodium bicarbonate solution for use in this protocol. Also note, because we adjust the pH of all 10X media and 10X balanced salt solution concentrates for solubility, you may need to adjust the pH after the dilution and to add sodium bicarbonate, as appropriate.
Media | Cat. No. | pH of 10X solution | NaHCO3 g/L | NaHCO3 mL/L 7.5% solution | pH after 1:10 dilution & NaHCO3 addition* | pH of 1X solution* |
---|---|---|---|---|---|---|
HBSS (10X), calcium, magnesium, phenol red | 14060040 | 5.5–6.0 | 0.35 | 4.7 | 7.5–7.8 | 7.0–7.2 |
HBSS (10X), calcium, magnesium, no phenol red | 14065049 14065056 14065072 | 4.4–4.7 | 0.35 | 4.7 | 7.5–7.8 | 7.0–7.4 |
DPBS (10X), calcium, magnesium | 1408004 14080055 14080089 | 4.4–4.7 | NA | NA | 5.2–5.5 | 7.0–7.2 |
HBSS (10X), no calcium, no magnesium, phenol red | 14180046 | 5.7–6.0 | 0.35 | 4.7 | 6.0–6.3 | 7.0–7.2 |
HBSS (10X), no calcium, no magnesium, no phenol red | 14185045 14185052 | 5.8–6.1 | 0.35 | 4.7 | 6.0–6.3 | 7.0–7.4 |
DPBS (10X), no calcium, no magnesium | 14200166 14200075 | 6.7–7.0 | NA | NA | 7.1–7.4 | 7.0–7.2 |
* pH range varies depending on the pH of water used in dilution.
Media sterilization reduces the risk of contamination by eliminating potential contaminants through autoclaving or manual filtration. In an autoclave, the media is subjected to high pressure and temperature, after which it must cool to room temperature before any supplements are added to prevent the degradation of heat-sensitive components (such as vitamins and amino acids). When media cannot be sterilized in an autoclave, such as when the media already contains heat-sensitive components, filtration is recommended. In the protocols above, a 0.2 μm filter system is used. This size is considered sterilizing grade and removes all bacteria and larger microbes.
Media supplementation involves adding essential nutrients and growth factors to the sterilized media to help provide an adequate environment for the cells. Supplements can introduce the proper pH balance, osmotic pressure, and essential nutrients to basal mediums. Common supplements include 7.5% sodium bicarbonate solution, L-glutamine, antibiotics, and serum.
Following proper storage and preparation procedures can help ensure success in routine cell culture.
Grow and maintain a variety of mammalian cell lines with Gibco Cell Culture Media.
Nunc Cell Culture Plastics support every step of your research with a broad range of culture vessels.
Gibco FBS offers consistent growth for your cells through essential nutrients and growth factors.
Access cell culture and transfection educational resources to enable excellent experiment planning and execution.
Learn best practices and fundamentals of cell culture. Topics include aseptic technique and methods for the growth and maintenance of cells in culture.
Explore this collection of cell culture videos providing valuable insights into techniques, resources, and stay up to date with the latest advancements in the field.
Find the right cell culture products for your research with these selection tools. Search for media, FBS, plastics, transfection reagents, and more.
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.