Epidemiological research
Epidemiological surveillance is conducted to ensure viral diseases match the reference strain and to monitor possible mutations, since any changes in the viral genome can impact public health policies and options, how the illness spreads in the population, potential study of treatment options, and vaccine development research. Thermo Fisher Scientific has a complete solution to assist scientists who are conducting epidemiological studies.
Virus typing
Identifying the types and subtypes of viruses is essential to good disease control. A wide variety of research methods can be used to differentiate virus variants of which molecular techniques are used most frequently. By sequencing a specific variable region of the viral genome, one can investigate whether selected isolates of a certain species are phylogenetically related. A major challenge for microbiologists and virologists is the prediction of patterns of evolution and emergence of disease agents. RNA viruses like SARS-CoV-2 (the coronavirus responsible for COVID-19) share the biological feature of high genetic variability, which causes them to appear as clouds of mutants. Advanced molecular technologies like next-generation sequencing (NGS) enables complete SARS-CoV-2 genome sequencing, including all variants and potential serotypes.
Viral typing with Ion Torrent NGS
Fast, automated and accurate targeted NGS workflow for SARS-CoV-2 epidemiological investigation
Targeted NGS is a rapid and accurate solution for viral genomes from a variety of epidemiological samples from viral investigations. Our viral typing NGS solutions provide ultra-high sensitivity, scalability, and rapid turnaround time, which are all vital to viral monitoring and investigation research. Ion Torrent targeted next-generation sequencing (NGS) enables a streamlined research workflow for complete genome sequencing and epidemiological studies of SARS-CoV-2.
Video Player is loading.
Current Time 0:00
Duration 3:34
Loaded: 0%
Stream Type LIVE
Remaining Time 3:34
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    The Ion AmpliSeq SARS-CoV-2 Insight Research Assay is designed to cover more than 99% of the SARS-CoV-2 genome, including all potential serotypes, and can be used for complete viral genome sequencing and variant detection.  The assay is part of a fast, automated, and accurate targeted NGS workflow that enables coronavirus typing in under a day. This end-to-end solution includes the assay and plug-in suite that was developed in collaboration with researchers at the forefront of the SARS-CoV-2 crisis.
    The Ion AmpliSeq SARS CoV-2 Insight Research Assay is now available for the Ion Torrent Genexus Integrated Sequencer. This new NGS solution enables you to rapidly go from nucleic acid to variant report in less than a single day with minimal hands-on time.
    Learn more about our portfolio of Ion Torrent NGS instrumentation for SAR-CoV-2 research or request project costs.
    Customer spotlight
    Triche_Timothy_06-18-10
    Select products for targeted sequencing workflow
    Several methods have been developed for the targeted enrichment of genomic DNA for a variety of clinical and research applications. Typically, they are based on a multiplexed PCR amplification reaction and DNA hybridization. All these DNA enrichment methods rely heavily on fragmentation of genomic DNA prior to amplification, resulting in relatively short (less than 1000 base-pair) sequencing templates.
    Targeted sequencing workflow implementing target captured by hybridization includes several general steps - sample preparation, library construction, target enrichment, sequencing and analysis.
    Catalog # Name Size Price (USD) Qty
    AM1980 GLOBINclear™-Human Kit, for globin mRNA depletion Each
    728.00
    15401029 Yeast tRNA, 50 mg Each
    278.65
    Online Exclusive
    307.00
    Save 28.35 (9%)
    AM1981 GLOBINclear™ Kit, mouse/rat, for globin mRNA depletion Each
    710.00
    AM8740 RNA Fragmentation Reagents Each
    123.00
    15279011 Human Cot-1 DNA™ Each
    355.65
    Online Exclusive
    391.00
    Save 35.35 (9%)
    18440016 Mouse Cot-1 DNA Each
    377.65
    Online Exclusive
    407.00
    Save 29.35 (7%)
    15279101 Human Cot-1 DNA™-Fluorometric QC Each
    609.65
    Online Exclusive
    662.00
    Save 52.35 (8%)
    19524016 Biotin-14-dATP Each
    644.65
    Online Exclusive
    712.00
    Save 67.35 (9%)
    19518018 Biotin-14-dCTP Each
    586.65
    Online Exclusive
    640.00
    Save 53.35 (8%)
    19519016 Biotin-14-CTP Each
    686.00
    18094011 BioPrime™ DNA Labeling System Each
    702.65
    Online Exclusive
    768.00
    Save 65.35 (9%)
    18095012 BioPrime™ Array CGH Genomic Labeling Module Each
    728.00
    18010025 DNA Polymerase I, 1000 U Each
    533.65
    Online Exclusive
    638.00
    Save 104.35 (16%)
    De novo sequencing using Ion Torrent NGS
    In addition to targeted sequencing, which covers >99% of the viral genome and variants, Ion Torrent systems can be used for hypothesis-free (de novo) genome sequencing of SARS-CoV-2 by NGS.
    ProductCat no.
    Ion AmpliSeq SARS-CoV-2 Insight Research AssayContact your sales representative
    1. Italian Researchers Identify New SARS-CoV-2 Gene Variants that Provide Clues to Coronavirus Epidemiology
    2. Molecular characterization of SARS-CoV-2 from the first case of COVID-19 in Italy- M.R. Capobianchi et al – National Institute for Infectious Diseases Lazzaro Spallanzani IRCCS, Rome, Italy
    3. High-density amplicon sequencing identifies community 1 spread and ongoing evolution of 2 SARS-CoV-2 in the Southern United States - Ryan P. McNamara et al - The University of North Carolina at Chapel Hill School of Medicine, Department of Microbiology and Immunology
    4.  Phylogenetic pattern of SARS-CoV-2 from COVID-19 patients from Bosnia and Herzegovina: lessons learned to optimize future molecular and epidemiological approaches - Teufik Goletic et al - Veterinary faculty of University of Sarajevo, Zmaja od Bosne 90, 71000 Sarajevo, Bosnia and Herzegovina
    Variant confirmation with Sanger sequencing
    While NGS analyses can be used for complete viral genome sequencing and variant detection of the SARS-CoV-2 genome, Sanger sequencing is still considered the gold standard sequencing method for verifying the sequence of specific genes, including those already sequenced through Illumina NGS due to its high degree of accuracy.  It is being used to sequence and confirm variants of different SARS-CoV-2 genes within clusters of symptomatic and asymptomatic individuals (1-4).
    The entire workflow can be completed, from sample to answer, in less than one workday.  We offer products to support the entire workflow from PCR amplification to data analysis.  The SeqStudio genetic analyzer is an affordable, low-throughput cartridge-based benchtop genetic analyzer that eliminates complex instrument set-up to deliver Sanger sequencing runs in as little as 30 minutes. 
    Learn more about Sanger sequencing solutions for SARS-CoV-2 research ›
    1.  Yuan, Y,. et al. (2020) “Molecular Epidemiology of SARS-CoV-2 Clusters Caused by Asymptomatic Cases in Anhui Province, China” Research Square preprint (https://dx.doi.org/10.21203/rs.3.rs-29833/v1)
    2.  Holland, LA., et al. (2020) “An 81 nucleotide deletion in SARS-1 CoV-2 ORF7a identified from sentinel surveillance in Arizona (Jan-Mar 2020)” J. Virology (https://doi.org/10.1128/jvi.00711-20)
    3.  Liu, Z., et al. (2020) “Identification of common deletions in 1 the spike protein of SARS-CoV-2”. J. Virology (https:// doi:10.1128/JVI.00790-20)
    4.  Tabibzadeh, A., et al. (2020) “SARS-CoV-2 Molecular and Phylogenetic analysis in COVID-19 patients: A preliminary report from Iran” Infection, Genetics and Evolution (https://doi.org/10.1016/j.meegid.2020.104387)
    Sanger Sequencing Solutions for SARS-CoV-2 Research
    Sanger_sequencing_SARS-CoV-2_research
    Generate highly specific targeted results
    Sanger sequencing can be completed in less than one workday—from sample to answer—helping to make your research of SARS-CoV-2 (the coronavirus that causes COVID-19) fast and cost-effective.  
    Download detailed protocols for single-sample sequencing of the virus to identify mutations and to confirm data from NGS sequencing.
    Learn More ›
    Virus monitoring
    Accurate understanding and monitoring of the SARS-CoV-2 virus is essential for effective response. Monitoring is important to help public health decision makers know where the disease is, how quickly it is spreading, and to longitudinally monitor the effectiveness of public health measures on outbreak containment or mitigation. Monitoring also plays a role in understanding cross-species transmission of the virus and its spatial spread over evolutionary time and at human-wildlife interfaces. We offer several cost-effective molecular tools for SARS-CoV-2 disease monitoring research, from technologies like real-time PCR and Sanger sequencing for specific genes/targets to whole genome sequencing to capture all variants using NGS.
    Targeted NGS workflow for SARS-CoV-2
    Targeted NGS is a rapid and accurate solution for viral genomes, suitable for monitoring SARS-CoV-2 infection and spread, including zoonotic transmission. Ion Torrent targeted NGS solutions provide ultra-high sensitivity, scalability, and rapid turnaround time, which are all vital to viral monitoring and investigation research. It enables a streamlined research workflow for complete genome sequencing and epidemiological studies of SARS-CoV-2.
    The Ion AmpliSeq SARS-CoV-2 Insight Research Assay is designed to cover more than 99% of the SARS-CoV-2 genome, including all potential serotypes, and can be used for complete viral genome sequencing and variant detection.  The assay is part of a fast, automated, and accurate targeted NGS workflow that enables coronavirus typing in under a day. This end-to-end solution includes the assay and plug-in suite that was developed in collaboration with researchers at the forefront of the SARS-CoV-2 crisis.
    Learn more about our portfolio of Ion Torrent NGS instrumentation for SAR-CoV-2 research or request project costs.
    1. Presence and vitality of SARS-CoV-2 virus in wastewaters and rivers - Sara Giordana Rimoldi et al - Laboratorio di Microbiologia Clinica, Virologia e Diagnostica delle Bioemergenze, ASST Fatebenefratelli Sacco, Milan, Italy;
    2. Lam, T.T., Jia, N., Zhang, Y. et al. Identifying SARS-CoV-2-related coronaviruses in Malayan pangolins. Nature583, 282–285 (2020). https://doi.org/10.1038/s41586-020-2169-0
    3. Molecular Detection of SARS-CoV-2 Infection in FFPE Samples and Histopathologic Findings in Fatal SARS-CoV-2 Cases - Miroslav Sekulic et al - Department of Pathology, University Hospitals Cleveland Medical Center, Cleveland, OH; Department of Pathology, Case Western Reserve University School of Medicine, Cleveland, OH
    SARS-CoV-2 monitoring using real-time PCR
    Real-time PCR can be used to research viral and human genetic determinants that influence the distribution and patterns of disease conditions within a population or large sample size. Real-time PCR testing takes less than 40 minutes, is accurate to <10-copy detection, and has an affordable cost per sample.
    SARS-CoV-2 has shown us that the time to and reproducibility of results are critical in potentially mitigating the impact of a crisis. A rigorous, reproducible workflow built on automation and integrated connectivity produces consistent results with fewer errors, while a coordinated ecosystem gives you complete control and oversight of your research. The following products used in concert with our suite of digital solutions enables you to connect everything: lab automation, data management, and digital partners, to help you move faster when timing is critical.
    Learn more about our real-time PCR solutions for SARS-CoV-2 research ›
    TaqMan Gene Expression Assays
    Catalog #Assay IDNameSize
    4331182Vi06439674_s1CoV_HKU1250 rxns
    4331182Vi06439673_s1CoV_NL63250 rxns
    4331182Vi06439671_s1CoV_229E250 rxns
    4331182Vi06439644_s1MERS_CoV250 rxns
    4331182Vi06439634_s1SARS_CoV250 rxns
    To order, search the Assay ID or Name using our ordering tool.    Ordering tool
    Other products
    Catalog # Name Size Price (USD) Qty
    A46110 PowerTrack™ SYBR Green Master Mix for qPCR, 2 x 5 mL Each
    935.65
    Online Exclusive
    1,002.00
    Save 66.35 (7%)
    SARS-CoV-2 monitoring with Sanger sequencing and fragment analysis by capillary electrophoresis
    Sanger sequencing is a simple, fast, and cost-effective orthogonal gold-standard method to monitor SARS-CoV-2 identified by NGS or by RT-PCR.  The entire workflow can be completed, from sample to answer, in less than one workday. It is being used by several labs globally to confirm the identity of SARS-CoV-2 in wastewater samples. It is also being used to confirm the origin and cross-species transmission of different coronaviruses, which is important in understanding potential sources of zoonotic transmission (1-3).
    Fragment analysis by capillary electrophoresis (CE) provides an attractive method for  rapid testing of multiple targets. Multiplexed qPCR solutions detect small numbers of pathogens. The relatively small capacity of multiplexed qPCR reactions can limit throughput when large numbers of targets or pathogens need to be detected. Fragment analysis gives researchers another tool for analyzing the presence of genomic sequences of pathogenic organisms using a method that is rapid, simple, and sensitive.
    Fragment analysis by CE can be used to identify microbial signatures associated with different disease phenotypes. It relies on amplification of 16S-23S rRNA interspace (IS) regions in bacteria to produce PCR fragments that vary in length and frequency depending upon bacterial species. Each PCR fragment can be separated and detected on a genetic analyzer. The resulting fragment patterns are compared to a curated database to determine the species present in the sample.
    1. Valitutto, M.T., et al. (2020) “Detection of novel coronaviruses in bats in Myanmar,” PLoS One 15(4): e0230802.( https://doi.org/10.1371/journal.pone.0230802)
    2. Latinne A., et al. (2020) Origin and cross-species transmission of bat coronaviruses in China. (bioRxiv preprint doi: https://doi.org/10.1101/2020.05.31.116061)
    3. Huong, N.Q., et al (2020) “Coronavirus testing indicates transmission risk increases along 2 wildlife supply chains for human consumption in Viet Nam, 2013-2014 (bioRxiv preprint doi: https://doi.org/10.1101/2020.06.05.098590.)
    SARS-CoV-2 Serology Surveillance
    New SARS-CoV-2 Serology Assays: Rapidly detect antibodies (immunoglobulins, IgG and IgM) in serum against the SARS-CoV-2 coronavirus.
    ELISA assay kit
    SARS-CoV-2 Ig ELISA
    This Human SARS-CoV-2 Spike (trimer) ELISA kit enables the detection of anti-SARS-CoV-2 Ig antibodies in human serum with great sensitivity and specificity.
    ProcartaPlex Assay kit
    ProcartaPlex SARS-Cov-2 Panel
    ProcartaPlex Human Coronavirus Ig Total 11-plex panel enables the detection of multiple anti-SARS-CoV-2 (Spike trimer, S1, RBD, N) Ig antibodies and 6 other coronavirus strains in human serum, helping to provide a more complete antibody profile.
    Sample Prep
    Viral nucleic acid isolation for SARS-CoV-2 research
    Prior to any NGS-based epidemiological study of SARS-CoV-2, researchers need to quickly and efficiently isolate RNA. Features of the MagMAX Viral/Pathogen Nucleic Acid Isolation Kit include:
    • Automation-ready protocols designed for Thermo Scientific KingFisher systems
    • Flexible protocols accommodate multiple sample types and input volumes
    • Recommended for viral nucleic acid isolation for SARS-CoV-2 to obtain high-quality nucleic acid
    For fast, easy, and effective high-throughput sample processing, automate your nucleic acid isolation with the KingFisher Purification systems, a versatile benchtop instrument for the processing of 24 or 96 samples per run. Features of this system include:
    • Easy to install and run (ready to run in 10 minutes or less)
    • Download existing protocols from a library, or easily customize protocols using Thermo Scientific BindIt Software
    Avoid high levels of contaminating DNA and RNases and maintain a nuclease-free lab with RNaseZap solutions and nuclease-free water, buffers and reagents, and tips and tubes products:
    Next-generation sequencing—library preparation for Illumina systems
    Collibri Library Prep Kits for Illumina Systems enhance next-generation sequencing (NGS)-based viral surveillance with higher quality sequences and rapid protocols. Full-length genomic coverage can improve the accuracy of monitored transmission patterns and mutation rates of SARS-CoV-2. Unlike transposomic methods, the Collibri DNA Library Prep Kits for Illumina Systems provide full-length, hypothesis-free, genomic coverage using a library prep method that is completed in 1.5 hours. Collibri DNA Library Prep Kits are suitable for genomes of all sizes.
    ProductCat no.
    GeneArt Gene Synthesis and Gene-to-Protein ServicesSee thermofisher.com/geneart
    cDNA synthesis for epidemiological research
    Invitrogen SuperScript reverse transcriptases were developed for improved thermostability, processivity, and cDNA yields to enable superior performance with even the most challenging RNA samples. With over 50,000 citations, reviews, and publications, SuperScript reverse transcriptases are the superior choice for vaccine development.
    See what other researchers are saying at www.thermofisher.com/ssiv
    Catalog # Name Size Price (USD) Qty
    18080044 SuperScript™ III Reverse Transcriptase, 10,000 Units Each
    481.65
    Online Exclusive
    528.00
    Save 46.35 (9%)
    12574026 SuperScript™ III One-Step RT-PCR System with Platinum™ Taq DNA Polymerase, 100 rxns Each
    944.65
    Online Exclusive
    1,060.00
    Save 115.35 (11%)
    12594100 SuperScript™ IV One-Step RT-PCR System, Each Each
    1,119.65
    Online Exclusive
    1,212.00
    Save 92.35 (8%)
    18090050 SuperScript™ IV Reverse Transcriptase, 10,000 Units Each
    497.65
    Online Exclusive
    530.00
    Save 32.35 (6%)
    For Research Use Only. Not for use in diagnostic procedures.