Cell Culture or tissue culture treated dish for adherent cells
Nunclon Delta Cell Culture treated plastics for adherent cells
Thermo Scientific Nunc cell culture–treated plastics with Nunclon Delta surface are proven for consistent cell growth. Every lot of Nunclon Delta surface plastics is validated with trusted Gibco media to confirm consistent cell growth across multiple cell lines.
Request sample/quote
What is Nunclon Delta?
Nunclon Delta is a cell culture–treated surface modification that makes the polystyrene surface of your culture vessel more hydrophilic, promoting maximum adhesion for a broad range of cell types.
This tissue-culture (TC) surface is offered across multiple formats for your adherent cell culture applications.
Thermo Scientific Nunc Cell Culture Treated Plates
Cell Culture or tissue culture treated multiwell plate for adherent cells
Promote maximum adhesion for a broad range of cell types with the Thermo Scientific Nunc Cell-Culture Treated multi-well plates with the Nunclon Delta surface treatment. The open format with the raised-well rim enables convenient and secure manipulation of the cells.
Available in 4- to 48-well multiwell platesand 96-well plates and 384 well microplates
Ordering information
Nunc 4–48 well cell culture treated plates
Catalog # Name Size Price (USD) Qty
176740 Nunc™ Cell-Culture Treated Multidishes, 4 well Case of 120
454.65
Online Exclusive
508.00
Save 53.35 (11%)
140675 6 Well Plate, TC Surface, Pack of 1 Case of 75
344.65
Online Exclusive
368.00
Save 23.35 (6%)
140685 6 Well Plate, TC Surface, Bulk Pack of 5 Case of 85
375.65
Online Exclusive
398.00
Save 22.35 (6%)
142475 24 Well Plate, TC Surface, Pack of 1 Case of 75
419.65
Online Exclusive
456.00
Save 36.35 (8%)
142485 24 Well Plate, TC Surface, Pack of 5 Case of 85
472.65
Online Exclusive
500.00
Save 27.35 (5%)
150628 12 Well Plate, TC Surface, Pack of 1 Case of 75
385.65
Online Exclusive
413.00
Save 27.35 (7%)
150687 48 Well Plate, TC Surface, Pack of 1 Case of 75
481.65
Online Exclusive
548.00
Save 66.35 (12%)
152640 48 Well Plate, TC Surface, Pack of 5 Case of 85
561.65
Online Exclusive
594.00
Save 32.35 (5%)
Nunc 96-and 384-well cell culture treated plates
Catalog # Name Size Price (USD) Qty
167008 96 Well Plate, TC Surface, Pack of 1 Case of 50
383.65
Online Exclusive
412.00
Save 28.35 (7%)
161093 96 Well Plate, TC Surface, Pack of 10 Case of 160
1,068.65
Online Exclusive
1,138.00
Save 69.35 (6%)
136101 96 Well White Plate, TC Surface, Pack of 1 Case of 50
402.65
Online Exclusive
424.00
Save 21.35 (5%)
136102 96 Well White Plate, TC Surface, Bulk 10 Pack Case of 160
1,202.65
Online Exclusive
1,266.00
Save 63.35 (5%)
137101 96 Well Black Plate, TC Surface, Pack of 1 Case of 50
377.65
Online Exclusive
424.00
Save 46.35 (11%)
137103 96 Well Black Plate, TC Surface, Pack of 10 Case of 160
1,165.65
Online Exclusive
1,308.00
Save 142.35 (11%)
168055 96 Well Plate, TC Surface, No Lid, Pack of 1 Case of 50
322.65
Online Exclusive
342.00
Save 19.35 (6%)
163320 96 Well Round (U) Bottom Plate, TC Surface, Pack of 1 Case of 50
383.65
Online Exclusive
410.00
Save 26.35 (6%)
168136 Nunc™ 96-Well, Nunclon Delta-Treated, U-Shaped-Bottom Microplate Case of 160
1,134.00
143761 96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1 Case of 50
294.00
164688 384 Well Plate, TC Surface, Pack of 10 Case of 30
280.65
Online Exclusive
293.00
Save 12.35 (4%)
Also try our Edge 2.0 plates to reduce plate evaporation
Catalog # Name Size Price (USD) Qty
167425 96 Well Edge Plate, TC Surface, Pack of 1 Case of 50
143.65
Online Exclusive
155.00
Save 11.35 (7%)
167542 96 Well Edge Plate, TC Surface, Pack of 10 Case of 160
424.65
Online Exclusive
459.00
Save 34.35 (7%)
167574 Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate Case of 50
138.65
Online Exclusive
147.00
Save 8.35 (6%)
167554 96 Well Edge Plate, TC Surface, No Lid, Pack of 10 Case of 160
395.00
Thermo Scientific Nunc EasYFlask Cell Culture Treated Flasks
Cell Culture or tissue culture treated flask for adherent cells
Enhance cell attachment, growth, and differentiation with EasYFlask cell culture flasks with Nunclon Delta surface treatment. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
Available in T25-T225
Ordering information
Catalog # Name Size Price (USD) Qty
156340 T25 EasYFlask, TC Surface, Solid Cap, Pack of 10 Case of 200
480.65
Online Exclusive
574.00
Save 93.35 (16%)
156367 T25 EasYFlask, TC Surface, Filter Cap, Pack of 10 Case of 200
605.65
Online Exclusive
662.00
Save 56.35 (9%)
156472 T75 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 100
493.65
Online Exclusive
540.00
Save 46.35 (9%)
156499 T75 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 100
491.65
Online Exclusive
560.00
Save 68.35 (12%)
159910 T175 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 30
286.65
Online Exclusive
327.00
Save 40.35 (12%)
159920 T175 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 30
227.65
Online Exclusive
318.00
Save 90.35 (28%)
159933 T225 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 30
288.65
Online Exclusive
405.00
Save 116.35 (29%)
159934 T225 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 30
328.65
Online Exclusive
393.00
Save 64.35 (16%)
Thermo Scientific Nunc EasYDish Cell Culture Treated Dishes
Cell Culture or tissue culture treated dish for adherent cells
Nunc EasYDish cell culture dishes are designed to improve handling, stacking, and transporting of cells with the help of a raised outer edge and grip ring.  With our Nunclon Delta treated surface treatment, these dishes are tested and certified for consistent cell growth across multiple cell lines.
Available in 35mm – 150 mm
Ordering information
Catalog # Name Size Price (USD) Qty
150460 35mm EasYDish, TC Surface, Pack of 10 Case of 500
433.65
Online Exclusive
474.00
Save 40.35 (9%)
150462 60mm EasYDish, TC Surface, Pack of 10 Case of 280
264.65
Online Exclusive
291.00
Save 26.35 (9%)
150464 100x17mm EasYDish, TC Surface, Pack of 10 Case of 150
306.65
Online Exclusive
336.00
Save 29.35 (9%)
150466 100x21mm EasYDish, TC Surface, Pack of 10 Case of 240
482.65
Online Exclusive
534.00
Save 51.35 (10%)
150468 150mm EasYDish, TC Surface, Pack of 10 Case of 80
340.65
Online Exclusive
372.00
Save 31.35 (8%)
Imaging cultureware
Thermo Scientific Nunc Cell Culture Imaging Products offer comprehensive solutions with flexibility and consistency to meet your needs in cell imaging applications.
Discover which formats fit your needs:
Cell Culture or Tissue culture treated black optical bottom plate for adherent cells
Optical bottom plates
With minimum background noise, the Thermo Scientific Nunc 96- and 384-well Optical Bottom Plates offer a wide selection of ideal for cell-based fluorescence and luminescence assays. Choose the best optical bottom plate for your application.
Cell Culture or tissue culture treated chamber slides for adherent cells
Chamber slides
Grow, fix, and stain your cells directly on Thermo Scientific Nunc Lab Tek Chamber Slides. Learn more about what chamber slide is right for you.
Ordering information
Optical bottom plates with Nunclon Delta Surface
Chamber slides with Nunclon Delta Surface
Catalog # Name Size Price (USD) Qty
177410 LabTek Chamber Slide, 1 Well Permanox TC Surface Case of 96
1,354.65
Online Exclusive
1,394.00
Save 39.35 (3%)
177429 LabTek Chamber Slide, 2 well Permanox TC Surface Case of 96
1,359.65
Online Exclusive
1,400.00
Save 40.35 (3%)
177437 LabTek Chamber Slide, 4 well Permanox TC Surface Case of 96
1,388.65
Online Exclusive
1,428.00
Save 39.35 (3%)
177445 LabTek Chamber Slide, 8-well Permanox TC Surface, Treated Case of 96
1,423.65
Online Exclusive
1,464.00
Save 40.35 (3%)
Thermo Scientific Nunc Cell Culture Treated Plates
Cell Culture or tissue culture treated multiwell plate for adherent cells
Promote maximum adhesion for a broad range of cell types with the Thermo Scientific Nunc Cell-Culture Treated multi-well plates with the Nunclon Delta surface treatment. The open format with the raised-well rim enables convenient and secure manipulation of the cells.
Available in 4- to 48-well multiwell platesand 96-well plates and 384 well microplates
Ordering information
Nunc 4–48 well cell culture treated plates
Catalog # Name Size Price (USD) Qty
176740 Nunc™ Cell-Culture Treated Multidishes, 4 well Case of 120
454.65
Online Exclusive
508.00
Save 53.35 (11%)
140675 6 Well Plate, TC Surface, Pack of 1 Case of 75
344.65
Online Exclusive
368.00
Save 23.35 (6%)
140685 6 Well Plate, TC Surface, Bulk Pack of 5 Case of 85
375.65
Online Exclusive
398.00
Save 22.35 (6%)
142475 24 Well Plate, TC Surface, Pack of 1 Case of 75
419.65
Online Exclusive
456.00
Save 36.35 (8%)
142485 24 Well Plate, TC Surface, Pack of 5 Case of 85
472.65
Online Exclusive
500.00
Save 27.35 (5%)
150628 12 Well Plate, TC Surface, Pack of 1 Case of 75
385.65
Online Exclusive
413.00
Save 27.35 (7%)
150687 48 Well Plate, TC Surface, Pack of 1 Case of 75
481.65
Online Exclusive
548.00
Save 66.35 (12%)
152640 48 Well Plate, TC Surface, Pack of 5 Case of 85
561.65
Online Exclusive
594.00
Save 32.35 (5%)
Nunc 96-and 384-well cell culture treated plates
Catalog # Name Size Price (USD) Qty
167008 96 Well Plate, TC Surface, Pack of 1 Case of 50
383.65
Online Exclusive
412.00
Save 28.35 (7%)
161093 96 Well Plate, TC Surface, Pack of 10 Case of 160
1,068.65
Online Exclusive
1,138.00
Save 69.35 (6%)
136101 96 Well White Plate, TC Surface, Pack of 1 Case of 50
402.65
Online Exclusive
424.00
Save 21.35 (5%)
136102 96 Well White Plate, TC Surface, Bulk 10 Pack Case of 160
1,202.65
Online Exclusive
1,266.00
Save 63.35 (5%)
137101 96 Well Black Plate, TC Surface, Pack of 1 Case of 50
377.65
Online Exclusive
424.00
Save 46.35 (11%)
137103 96 Well Black Plate, TC Surface, Pack of 10 Case of 160
1,165.65
Online Exclusive
1,308.00
Save 142.35 (11%)
168055 96 Well Plate, TC Surface, No Lid, Pack of 1 Case of 50
322.65
Online Exclusive
342.00
Save 19.35 (6%)
163320 96 Well Round (U) Bottom Plate, TC Surface, Pack of 1 Case of 50
383.65
Online Exclusive
410.00
Save 26.35 (6%)
168136 Nunc™ 96-Well, Nunclon Delta-Treated, U-Shaped-Bottom Microplate Case of 160
1,134.00
143761 96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1 Case of 50
294.00
164688 384 Well Plate, TC Surface, Pack of 10 Case of 30
280.65
Online Exclusive
293.00
Save 12.35 (4%)
Also try our Edge 2.0 plates to reduce plate evaporation
Catalog # Name Size Price (USD) Qty
167425 96 Well Edge Plate, TC Surface, Pack of 1 Case of 50
143.65
Online Exclusive
155.00
Save 11.35 (7%)
167542 96 Well Edge Plate, TC Surface, Pack of 10 Case of 160
424.65
Online Exclusive
459.00
Save 34.35 (7%)
167574 Nunc™ Edge™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate Case of 50
138.65
Online Exclusive
147.00
Save 8.35 (6%)
167554 96 Well Edge Plate, TC Surface, No Lid, Pack of 10 Case of 160
395.00
Thermo Scientific Nunc EasYFlask Cell Culture Treated Flasks
Cell Culture or tissue culture treated flask for adherent cells
Enhance cell attachment, growth, and differentiation with EasYFlask cell culture flasks with Nunclon Delta surface treatment. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
Available in T25-T225
Ordering information
Catalog # Name Size Price (USD) Qty
156340 T25 EasYFlask, TC Surface, Solid Cap, Pack of 10 Case of 200
480.65
Online Exclusive
574.00
Save 93.35 (16%)
156367 T25 EasYFlask, TC Surface, Filter Cap, Pack of 10 Case of 200
605.65
Online Exclusive
662.00
Save 56.35 (9%)
156472 T75 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 100
493.65
Online Exclusive
540.00
Save 46.35 (9%)
156499 T75 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 100
491.65
Online Exclusive
560.00
Save 68.35 (12%)
159910 T175 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 30
286.65
Online Exclusive
327.00
Save 40.35 (12%)
159920 T175 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 30
227.65
Online Exclusive
318.00
Save 90.35 (28%)
159933 T225 EasYFlask, TC Surface, Solid Cap, Pack of 5 Case of 30
288.65
Online Exclusive
405.00
Save 116.35 (29%)
159934 T225 EasYFlask, TC Surface, Filter Cap, Pack of 5 Case of 30
328.65
Online Exclusive
393.00
Save 64.35 (16%)
Thermo Scientific Nunc EasYDish Cell Culture Treated Dishes
Cell Culture or tissue culture treated dish for adherent cells
Nunc EasYDish cell culture dishes are designed to improve handling, stacking, and transporting of cells with the help of a raised outer edge and grip ring.  With our Nunclon Delta treated surface treatment, these dishes are tested and certified for consistent cell growth across multiple cell lines.
Available in 35mm – 150 mm
Ordering information
Catalog # Name Size Price (USD) Qty
150460 35mm EasYDish, TC Surface, Pack of 10 Case of 500
433.65
Online Exclusive
474.00
Save 40.35 (9%)
150462 60mm EasYDish, TC Surface, Pack of 10 Case of 280
264.65
Online Exclusive
291.00
Save 26.35 (9%)
150464 100x17mm EasYDish, TC Surface, Pack of 10 Case of 150
306.65
Online Exclusive
336.00
Save 29.35 (9%)
150466 100x21mm EasYDish, TC Surface, Pack of 10 Case of 240
482.65
Online Exclusive
534.00
Save 51.35 (10%)
150468 150mm EasYDish, TC Surface, Pack of 10 Case of 80
340.65
Online Exclusive
372.00
Save 31.35 (8%)
Imaging cultureware
Thermo Scientific Nunc Cell Culture Imaging Products offer comprehensive solutions with flexibility and consistency to meet your needs in cell imaging applications.
Discover which formats fit your needs:
Cell Culture or Tissue culture treated black optical bottom plate for adherent cells
Optical bottom plates
With minimum background noise, the Thermo Scientific Nunc 96- and 384-well Optical Bottom Plates offer a wide selection of ideal for cell-based fluorescence and luminescence assays. Choose the best optical bottom plate for your application.
Cell Culture or tissue culture treated chamber slides for adherent cells
Chamber slides
Grow, fix, and stain your cells directly on Thermo Scientific Nunc Lab Tek Chamber Slides. Learn more about what chamber slide is right for you.
Ordering information
Optical bottom plates with Nunclon Delta Surface
Chamber slides with Nunclon Delta Surface
Catalog # Name Size Price (USD) Qty
177410 LabTek Chamber Slide, 1 Well Permanox TC Surface Case of 96
1,354.65
Online Exclusive
1,394.00
Save 39.35 (3%)
177429 LabTek Chamber Slide, 2 well Permanox TC Surface Case of 96
1,359.65
Online Exclusive
1,400.00
Save 40.35 (3%)
177437 LabTek Chamber Slide, 4 well Permanox TC Surface Case of 96
1,388.65
Online Exclusive
1,428.00
Save 39.35 (3%)
177445 LabTek Chamber Slide, 8-well Permanox TC Surface, Treated Case of 96
1,423.65
Online Exclusive
1,464.00
Save 40.35 (3%)
Nunc cell culture–treated plastics are tested with Gibco Media for optimal adherent cell growth
Healthy adherent cell growth requires quality treated plastics. Only Thermo Scientific cell culture plastics endure rigorous testing with Gibco media, including cloning efficiency tests using specially selected cell lines with high sensitivity to toxic components. It’s the perfect pairing that can mean the difference between success and starting over.
Thermo Scientific Nunc flasks, dishes, and plates feature the standard Nunclon Delta cell culture–treated surface for growing adherent cells. Every lot of these TC products is tested and certified for cell growth and plating efficiency using 4 different cell types: L-929, HEL 299, V79-4, and primary chick embryo cells.
Every lot of Nunclon Delta TC treated plastics are tested across adherent cell lines L-929, HEL 299, and primary chick embryo cells for cell growth and plating efficiency and for V79-4 cloning efficiency.
Nunclon Delta certification
For the L-929, HEL 299, and primary chick embryo cells, growth is assessed by the percentage of surface coverage per cultureware. The growth of V79-4 cells is evaluated by the number of colonies formed per cultureware. For Nunclon Delta lot certification, the Nunc cultureware has to demonstrate:
  • Consistent cell growth over the entire culture surface
  • Less than a 10% difference from the Nunclon Delta–qualified standards for L-929, HEL 299, and primary chick embryo cell growth
  • Less than a 15% difference from the Nunclon Delta–qualified standards for V79-4 colony formation
Learn about our cell culture testing for Nunclon Delta certification
Nunclon Delta resources
Gibco and Nunc are Better Together
Nunc cell culture plastics, Gibco media and FBS are a proven combination for happy cells—and happy scientists.  Discover how these products are pairs for productivity and convenience. Learn more
Nunclon Surface Cell Line Data Tool
Get to know the facts about your cell culture–treated plastics by using our new Nunclon Surface Cell Line Data tool. We've tested the most commonly used cell lines on TC treated Thermo Scientific Nunclon Delta surface side-by-side with the leading competitor to validate Nunclon Delta's place in your cell culture hood. Use the Nunclon Surface Cell Line Data tool
Not using Thermo Scientific Nunc Cell Culture plastics?
Switch to Nunc cell culture–treated plastics with help of our cross-reference tool. Enter your supplier's product number (SKU) into the search field within the tool to find the Nunc equivalent or filter products by brand, category, and size. Find Nunc Cell Culture Plastics now
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products
For Research Use Only. Not for use in diagnostic procedures.