Diagram of Grignard reactions
Organomagnesium halides of the general structure R-Mg-X, with R = alkyl, alkenyl or aryl and X = halogen, are called “Grignard” reagents after their inventor, chemist Victor Grignard. Grignard reagents are highly reactive and react with most organic compounds. It also reacts with water, carbon dioxide, and oxygen. Grignard reagents are prepared by the reaction of magnesium metal with appropriate alkyl halide in ether solvent. The halogen may be chlorine, bromine, or iodine. A related synthesis uses ethylene oxide to prepare alcohols containing two more carbon atoms than that of the alkyl halide. Perhaps the most important use of the Grignard reagent is the reaction with aldehydes and ketones to form alcohols.
Thermo Fisher Scientific offers a range of Grignard reagents and related products in a variety of formats, including custom and bulk quantities.
Review available Thermo Scientific products for the Grignard reaction
Catalog # Name Price (USD) Qty
H51159.AD 1-Naphthylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
260.00
H51159.AE 1-Naphthylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
414.00
H54282.AE 2,4,6-Trimethylphenylmagnesium bromide, 1M in 2-MeTHF, Thermo Scientific Chemicals
385.00
H54159.AE 2,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
316.65
Online exclusive
352.00
Save 35.35 (10%)
H54966.AE 2,4-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
305.65
Special offer
Online exclusive
Ends: 30-Jun-2025
359.00
Save 53.35 (15%)
H54337.AE 2,5-Dimethoxyphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
248.65
Online exclusive
276.00
Save 27.35 (10%)
H54189.AE 2,6-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
233.65
Online exclusive
259.00
Save 25.35 (10%)
H54138.AE 2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
133.00
H51171.AE 2-Butenylmagnesium chloride, 0.5M in MeTHF, Thermo Scientific Chemicals
222.65
Special offer
Online exclusive
Ends: 26-Sep-2025
262.00
Save 39.35 (15%)
H54820.AE 2-Chlorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
281.00
H54881.AE 2-Fluorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals -
H54177.AE 2-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
536.65
Online exclusive
596.00
Save 59.35 (10%)
H54133.AE 3,4-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals -
H54312.AE 3,4-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
246.65
Online exclusive
274.00
Save 27.35 (10%)
H54197.AE 3,4-Difluorophenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
258.00
H54654.AE 3,4-Dimethoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals
250.65
Special offer
Online exclusive
Ends: 26-Sep-2025
295.00
Save 44.35 (15%)
H54632.AE 3,4-(Methylenedioxy)phenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
242.00
H54427.AE 3,5-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
204.65
Special offer
Online exclusive
Ends: 26-Sep-2025
241.00
Save 36.35 (15%)
H54622.AE 3,5-Difluorophenylmagnesium bromide, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
231.00
H54781.AE 3,5-Dimethyl-4-methoxyphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals -
H54057.AE 3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
446.65
Online exclusive
496.00
Save 49.35 (10%)
H54075.AE 3-Chloro-4-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
185.65
Special offer
Online exclusive
Ends: 30-Jun-2025
218.00
Save 32.35 (15%)
H54237.AE 3-Chlorobenzylmagnesium chloride, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
296.65
Special offer
Online exclusive
Ends: 30-Jun-2025
349.00
Save 52.35 (15%)
H51161.AD 3-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals
226.65
Online exclusive
252.00
Save 25.35 (10%)
H51161.AE 3-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals
374.65
Special offer
Online exclusive
Ends: 30-Jun-2025
441.00
Save 66.35 (15%)
H54335.AE 3-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals -
H54780.AE 3-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF, Thermo Scientific Chemicals
148.00
H54115.AE 4-(2-Tetrahydropyranyloxy)phenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
422.65
Online exclusive
469.00
Save 46.35 (10%)
H54625.AE 4-Chlorobenzylmagnesium chloride, 0.50M in 2-Me-THF, Thermo Scientific Chemicals -
H51163.AE 4-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals
64.65
Online exclusive
71.80
Save 7.15 (10%)
H54865.AE 4-Fluoro-2-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
231.65
Special offer
Online exclusive
Ends: 30-Jun-2025
272.00
Save 40.35 (15%)
H54201.AE 4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
213.65
Special offer
Online exclusive
Ends: 30-Jun-2025
251.00
Save 37.35 (15%)
H54078.AE 4-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals
350.65
Special offer
Online exclusive
Ends: 30-Jun-2025
412.00
Save 61.35 (15%)
089435.AE 4-Methoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals
79.65
Special offer
Online exclusive
Ends: 30-Jun-2025
94.10
Save 14.45 (15%)
089435.AP 4-Methoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals
309.00
H54317.AE 4-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF, Thermo Scientific Chemicals
143.65
Online exclusive
159.00
Save 15.35 (10%)
H54411.AE 4-Phenoxyphenylmagnesium bromide, 0.5 M in 2-MeTHF, Thermo Scientific Chemicals -
H54284.AE 4-tert-Butylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals
286.65
Special offer
Online exclusive
Ends: 30-Jun-2025
337.00
Save 50.35 (15%)
H54501.AE 5-Chloro-2-methoxyphenylmagnesium bromide, 0.50M in 2-MeTHF, Thermo Scientific Chemicals
212.65
Special offer
Online exclusive
Ends: 26-Sep-2025
250.00
Save 37.35 (15%)
H51170.AE Allylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
130.00
H51170.AP Allylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
449.65
Special offer
Online exclusive
Ends: 30-Jun-2025
529.00
Save 79.35 (15%)
087299.AD Benzylmagnesium chloride, 1-2M in THF, Thermo Scientific Chemicals
83.30
H51154.AE Benzylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
253.65
Online exclusive
282.00
Save 28.35 (10%)
H51154.AP Benzylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
946.00
H51165.AE Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
55.60
H51165.AP Cyclohexylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
186.65
Special offer
Online exclusive
Ends: 30-Jun-2025
219.00
Save 32.35 (15%)
H51158.AD Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
192.65
Online exclusive
214.00
Save 21.35 (10%)
H51158.AE Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals
301.65
Online exclusive
335.00
Save 33.35 (10%)
H26273.AE Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
232.65
Special offer
Online exclusive
Ends: 26-Sep-2025
274.00
Save 41.35 (15%)
087291.JT Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
69.10
087291.JU Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
108.00
041675.JT Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
65.00
041675.JU Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
122.00
H54840.AE Isopropylmagnesium bromide, 3M in 2-MeTHF, Thermo Scientific Chemicals
314.65
Special offer
Online exclusive
Ends: 26-Sep-2025
370.00
Save 55.35 (15%)
H51155.AE Isopropylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
127.65
Special offer
Online exclusive
Ends: 26-Sep-2025
150.00
Save 22.35 (15%)
H51155.AP Isopropylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
525.65
Online exclusive
584.00
Save 58.35 (10%)
H51156.AE Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF, Thermo Scientific Chemicals
252.65
Online exclusive
280.00
Save 27.35 (10%)
H51156.AP Isopropylmagnesium chloride - LiCl complex, 1M in MeTHF, Thermo Scientific Chemicals
997.65
Online exclusive
1,108.00
Save 110.35 (10%)
087324.AP Methylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
141.65
Special offer
Online exclusive
Ends: 26-Sep-2025
166.00
Save 24.35 (15%)
041252.AE Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
43.65
Special offer
Online exclusive
Ends: 30-Jun-2025
50.60
Save 6.95 (14%)
041252.AP Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
154.65
Online exclusive
172.00
Save 17.35 (10%)
042101.JS Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals -
042101.JU Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals -
H51162.AE n-Propylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
67.20
H51162.AP n-Propylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
213.65
Special offer
Online exclusive
Ends: 26-Sep-2025
251.00
Save 37.35 (15%)
H54729.AE Phenylmagnesium bromide, 3M in 2-MeTHF, Thermo Scientific Chemicals
274.65
Special offer
Online exclusive
Ends: 30-Jun-2025
323.00
Save 48.35 (15%)
087326.AE Phenylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
65.10
087326.AP Phenylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
163.65
Special offer
Online exclusive
Ends: 30-Jun-2025
192.00
Save 28.35 (15%)
042859.AE Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
62.20
042859.AP Phenylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
173.65
Online exclusive
193.00
Save 19.35 (10%)
H51164.AE tert-Butylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
151.65
Special offer
Online exclusive
Ends: 30-Jun-2025
178.00
Save 26.35 (15%)
H51164.AP tert-Butylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
682.65
Special offer
Online exclusive
Ends: 30-Jun-2025
803.00
Save 120.35 (15%)
H54824.AE tert-Pentylmagnesium chloride, 1M in 2-MeTHF, Thermo Scientific Chemicals
373.65
Special offer
Online exclusive
Ends: 26-Sep-2025
439.00
Save 65.35 (15%)
H54635.AE (Trimethylsilyl)methylmagnesium chloride, 0.5M in 2-MeTHF, Thermo Scientific Chemicals
136.65
Online exclusive
152.00
Save 15.35 (10%)