KingFisher plastics
KingFisher plastics are specifically designed for use with KingFisher instruments and are carefully manufactured to exacting standards.* Their low binding affinity for biomolecules makes them ideal for magnetic particle processing. The disposable polypropylene tip combs and microplates enable excellent recovery of magnetic beads for optimal purification and isolation of DNA, RNA, proteins, and cells.
All KingFisher plastics are made in a controlled environment that conforms to ISO 14644 Class ISO 8 (equivalent to US Federal Standard 209E Class 100,000), where the maximum number of particles permitted per cubic meter (m3) of air is ≥0.5 μm 3,520,000 and ≥5 μm 29,300.
KingFisher plastics
* Use of KingFisher plastics ensures proper KingFisher instrument function and application performance and avoids costly repairs that may be caused by using third-party plastics, even if marketed as “KingFisher compatible.” Use of third-party plastics may void instrument service contracts.
KingFisher Flex
KingFisher Flex plastics for 96 deep-well format
KingFisher Flex plastics for 96 standard and PCR formats
Catalog # Name Size Price (USD) Qty
97002540 Case of 48
202.65
Online Exclusive
257.00
Save 54.35 (21%)
97002524 Case of 100
682.00
97002514 Case of 80
480.65
Online Exclusive
556.00
Save 75.35 (14%)
KingFisher Flex plastics for 24 deep-well format
Catalog # Name Size Price (USD) Qty
95040470 Case of 50
317.65
Online Exclusive
395.00
Save 77.35 (20%)
95040480 Case of 50
414.65
Online Exclusive
491.00
Save 76.35 (16%)
97002610 Case of 50
899.65
Online Exclusive
1,164.00
Save 264.35 (23%)
97002620 Case of 50
1,040.65
Online Exclusive
1,268.00
Save 227.35 (18%)
KingFisher Presto
KingFisher Presto plastics for 96 deep-well format
Catalog # Name Size Price (USD) Qty
95040450 Case of 50
337.65
Online Exclusive
445.00
Save 107.35 (24%)
95040460 Case of 50
534.00
97002534 Case of 100
695.65
Online Exclusive
880.00
Save 184.35 (21%)
97002820 Case of 50
863.65
Online Exclusive
1,004.00
Save 140.35 (14%)
KingFisher Presto plastics for 96 standard and PCR formats
Catalog # Name Size Price (USD) Qty
97002540 Case of 48
202.65
Online Exclusive
257.00
Save 54.35 (21%)
KingFisher Presto plastics for 24 deep-well format
Catalog # Name Size Price (USD) Qty
95040470 Case of 50
317.65
Online Exclusive
395.00
Save 77.35 (20%)
95040480 Case of 50
414.65
Online Exclusive
491.00
Save 76.35 (16%)
97002610 Case of 50
899.65
Online Exclusive
1,164.00
Save 264.35 (23%)
97002620 Case of 50
1,040.65
Online Exclusive
1,268.00
Save 227.35 (18%)
KingFisher Duo Prime
KingFisher Duo Prime plastics for 96 deep-well format
KingFisher Duo Prime plastics for 24 deep-well format
Catalog # Name Size Price (USD) Qty
95040470 Case of 50
317.65
Online Exclusive
395.00
Save 77.35 (20%)
95040480 Case of 50
414.65
Online Exclusive
491.00
Save 76.35 (16%)
97003510 Each
197.65
Online Exclusive
242.00
Save 44.35 (18%)
KingFisher Apex
KingFisher Apex 96 Deep-Well Plates—15 to 1,000 µL
Plastics for 96 format
Catalog # Name Size Price (USD) Qty
AB2396 Each
124.65
Online Exclusive
197.00
Save 72.35 (37%)
A31728 Each
329.65
Online Exclusive
366.00
Save 36.35 (10%)
AB2496 Each
193.00
A36930 Each
142.00
4346907 Each
63.75
97002570 Case of 100
718.65
Online Exclusive
840.00
Save 121.35 (14%)
95040450B Case of 50
385.65
Online Exclusive
448.00
Save 62.35 (14%)
97002534B Case of 100
760.65
Online Exclusive
882.00
Save 121.35 (14%)
374086 Case of 960
1,330.00
Online Exclusive
1,400.00
Save 70.00 (5%)
97002560 Case of 50
351.65
Online Exclusive
407.00
Save 55.35 (14%)
97002540B Case of 48
221.65
Online Exclusive
258.00
Save 36.35 (14%)
97002524B Case of 100
549.65
Online Exclusive
700.00
Save 150.35 (21%)
95040460B Case of 50
454.65
Online Exclusive
526.00
Save 71.35 (14%)
97002820B Case of 50
980.00
3744 Case of 480
542.30
Online Exclusive
638.00
Save 95.70 (15%)
KingFisher Apex 24 Deep-Well Plates—30 to 5,000 µL
Plastics for 24 format
Catalog # Name Size Price (USD) Qty
97002580 Case of 50
861.65
Online Exclusive
1,066.00
Save 204.35 (19%)
95040470B Case of 50
341.65
Online Exclusive
406.00
Save 64.35 (16%)
97002610B Case of 50
954.65
Online Exclusive
1,174.00
Save 219.35 (19%)
95040480B Case of 50
476.00
97002620B Case of 50
1,078.65
Online Exclusive
1,234.00
Save 155.35 (13%)
374323 Case of 240
440.10
Online Exclusive
489.00
Save 48.90 (10%)
KingFisher Apex ordering information
Accessories ordering information
Additional accessories are available for the KingFisher system. KingFisher is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-processing for DNA/RNA, protein, and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality.
KingFisher Magnetic heads
KingFisher Apex accessories
KingFisher Flex accessories
Plate setup
To save time and increase reproducibility with your KingFisher system, combine it with an electronic pipette or automated reagent dispenser.
E1-ClipTip Equalizer Pipette
With the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette, you can perform sample transfers quickly and efficiently between virtually any tube, rack, microplate, or horizontal gel box. Adjustable tip spacing allows you to set the distance between tips simply by sliding the scale to expand or contract to the desired labware format. The new My Pipette™ Creator app lets you program the E1-ClipTip conveniently from your PC.
E1-ClipTip
Catalog # Name Size Price (USD) Qty
4672060BT Each
2,489.40
Online Exclusive
2,766.00
Save 276.60 (10%)
94420153 Case of 3840
567.86
Online Exclusive
636.00
Save 68.14 (11%)
* Other models that handle different sample volumes and tip sizes are available by following the Cat. No. links.
Multidrop Combi Reagent Dispenser
Fill your plates faster and more consistently with the MultiDrop™ Combi Reagent Dispenser, which provides precise dispensing over a wide 0.5 to 2500 µL range and accommodates microplates from 6 to 1,536 wells and plate heights of 5 to 50 mm.
Multidrop Combi Reagent Dispenser
Laboratory workflow automation
Streamline your KingFisher system workflow with a compatible barcode reader, robotic liquid handler, or robotic plate mover.
Robotic liquid handlers
The versatile KingFisher Presto is designed to integrate with a compatible robotic liquid handler for hands-free automation. Many commercial robotic liquid handlers are compatible with KingFisher Presto. Contact your KingFisher sales representative or your preferred vendor to see if a tested KingFisher interface is available.
Orbitor RS2 Microplate Mover
You can increase instrument loading throughput and reduce hands-on time with the Thermo Scientific™ Orbitor™ Microplate Mover, a compact and collaborative benchtop plate mover with integrated barcode scanning for sample tracking and inventory management. The Orbitor RS2’s innovative bidirectional telescoping arm provides exceptional reach and precision while the 360º workspace makes it a valuable tool for the lab. Compatible with the KingFisher Flex and Presto models, the Orbitor RS2 can be flexibly configured to match your requirements.
Thermo Scientific Orbitor RS2 Microplate Mover
Catalog # Name Size Price (USD) Qty
ORB2001 Each -
* Other configurations are available by following the Cat. No. link.
NanoDrop microvolume spectrophotometers
Quantify DNA, RNA, and protein samples in seconds with only 1–2 µL using Thermo Scientific NanoDrop microvolume spectrophotometers. You can rely on the proven technology in NanoDrop instruments for accurate, fast assessments of sample concentration and purity. By detecting and analyzing the whole spectrum, Thermo Scientific Acclaro sample intelligence software can help find and correct contamination in your samples and help differentiate DNA from RNA.
High-throughput SARS-CoV-2 testing solution
The Thermo Fisher Scientific Amplitude Solution is a new, highly automated molecular diagnostic testing system that can analyze over 7,000 COVID-19 specimens in 24 hours. The Amplitude Testing Solution can help laboratories quickly scale COVID-19 testing to volumes needed to help reduce the spread of disease and restore economies and communities. This solution offers maximum throughput with minimal hands-on time, equipment, and staffing—only four employees are required for operation.
As the world leader in serving science, Thermo Fisher Scientific has brought together industry-leading products like the KingFisher Presto and the Applied Biosystems TaqPath COVID-19 Kit into a high-throughput diagnostic solution. It offers the convenience of purchasing a secured supply of assays and consumables directly from a single-source supplier. Your initial purchase comes with an up-front monthly delivery of reagents for your laboratory’s needs over the first year. Our all-inclusive service partnership will handle every detail of the process to ensure a seamless installation and offers 24/7, 365-day priority access to reliable experts you can count on for any issues that arise now and into the future.
1x1 image pixel for data collection