Protein Electrophoresis Buffers and Reagents bottle
Finding the right protein gel electrophoresis system is critical to getting consistent and accurate results you need for your research. Electrophoresis buffers and reagents are important components of the protein electrophoresis system. We offer a range of SDS-PAGE buffers, native buffers and reagents for gel casting, sample preparation, running, and transferring gels. Find the recommended electrophoresis buffers and reagents for each gel system below.
Protein gels  Chamber systems
SDS sample buffers or loading buffers
Protein samples prepared for SDS-PAGE analysis are denatured by heating in the presence of a sample buffer containing 1–2% SDS or LDS with or without a reducing agent such as 20 mM DTT, 2-mercaptoethanol (BME) or TCEP. Sample buffers or loading buffers also contain glycerol so that they are heavier than water and sink neatly to the bottom of the buffer-submerged well when added to a gel. In addition, a negatively charged, low-molecular weight dye is also included in the sample buffer that will migrate at the buffer-front, enabling one to monitor the progress of electrophoresis. The most common tracking dyes for sample loading buffers are bromophenol blue, phenol red, and Coomassie blue. The table below summaries common sample buffers used in the different gel buffer systems.
Gel systemDenaturing sample buffer (final concentrations)Sample preparation
Tris-glycineTris-glycine SDS sample buffer: Tris HCl (63 mM), glycerol (10%), SDS (2%), bromophenol blue (0.0025%), pH 6.8Heat samples at 85°C for 2–5 minutes with sample buffer
Bis-TrisLDS sample buffer: Tris base (141 mM), Tris HCl (106 mM), LDS (2%), EDTA (0.51 mM), SERVA Blue G-250 (0.22 mM), phenol red (0.175 mM), pH 8.5Heat samples at 70°C for 10 minutes with sample buffer
Tris-AcetateLDS sample buffer: Tris base (141 mM), Tris HCl (106 mM), LDS (2%), EDTA (0.51 mM), SERVA Blue G-250 (0.22 mM), phenol red (0.175 mM), pH 8.5Heat samples at 70°C for 10 minutes with sample buffer
Tris-TricineTricine SDS sample buffer: Tris HCl (450 mM), glycerol (12%), SDS (4%), Coomassie Blue G (0.00075%), phenol red (0.0025%), pH 8.45Heat samples at 85°C for 2–5 minutes with sample buffer
ZymogramTris-glycine SDS: Tris HCl (63 mM), glycerol (10%), SDS (2%), bromophenol blue (0.0025%), pH 6.8Do not heat samples, load immediately onto gel after adding to sample
For fluorescence applications, we recommend that you use sample buffers that do not contain dyes that interfere in fluorescent channels (e.g., bromophenol blue). Fluorescent Compatible Sample Buffer does not contain bromophenol blue or other interfering dyes and can be used with reducing or nonreducing SDS-PAGE applications.
Find SDS sample buffer recipes in gel-specific product manuals in the documents tab.
Native sample buffers
Protein samples prepared for native PAGE applications are prepared in sample buffers or loading buffers that do not contain detergents to maintain the native state of the proteins. The loading buffers contain glycerol so that they are heavier than water and sink neatly to the bottom of the buffer-submerged well when added to a gel. In addition, a negatively charged, low-molecular weight dye is also included in the sample buffer that will migrate at the buffer-front, enabling one to monitor the progress of electrophoresis.
When preparing samples for native applications, protein samples should not be heated and loaded immediately onto the gel after mixing with the sample buffer.
Gel systemNative sample buffer (final concentrations)
Tris-glycineNative sample buffer: Tris HCl (100 mM), glycerol (10%), bromophenol blue (0.00025%), pH 8.6
Tris-acetateNative sample buffer: Tris HCl (100 mM), glycerol (10%), bromophenol blue (0.00025%), pH 8.6
IEFIEF sample buffer pH 3-7: Lysine (40 mM), glycerol (15%)
IEF sample buffer pH 3-10: Arginine (20 mM), Lysine (20 mM), glycerol (15%)
NativePAGE Bis-TrisNativePAGE Sample Buffer (4X): Bis-Tris (50 mM), 6 N HCl, NaCl (50 mM), Glycerol (10%), Ponceau S (0.001%), pH 7.2
Find native sample buffer recipes in gel-specific product manuals in the documents tab.
SDS running buffers
In protein electrophoresis (discontinuous buffer system), the primary anion in the gel is different (or discontinuous) from the primary anion in the running buffer. The tables below have the recommended SDS running buffers or native running buffers for each of the different gel chemistry buffering systems.
Gel systemSDS running buffer
Tris-glycineTris-glycine SDS: Tris base (25 mM), glycine (192 mM), SDS (0.1%), pH 8.3
Bis-TrisMES SDS: MES (50 mM), Tris base (50 mM), SDS (0.1%), EDTA (1 mM), pH 7.3
MOPS SDS: MOPS (50 mM), Tris base (50 mM), SDS (0.1%), EDTA (1 mM), pH 7.7
Tris-acetateTris-acetate SDS: Tris base (50 mM), Tricine (50 mM), SDS (0.1%), pH 8.24
Tris-tricineTricine-SDS: Tris base (100 mM), tricine (100 mM), SDS (0.1%), pH 8.3
Native running buffers
Gel systemNative running buffer
Tris-glycineTris-glycine native buffer: Tris base (25 mM), glycine (192 mM), pH 8.3
Tris-acetateTris-glycine native buffer: Tris base (25 mM), glycine (192 mM), pH 8.3
IEFIEF cathode buffer pH 3-7: Lysine (40 mM)
IEF cathode buffer pH 3-10: Arginine (20 mM), lysine (20 mM)
IEF anode buffer: phosphoric acid 85% (7 mM)
ZymogramTris-glycine SDS: Tris base (25 mM), glycine (192 mM), SDS (0.1%), pH 8.3
Find SDS running buffer recipes and native running buffer recipes in gel specific product manuals in the documents tab.
NuPAGE Bis-Tris SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Bis-trisDenaturing: LDS sample buffer
Reducing: Sample reducing agent
Small to medium-sized proteins (6–260 kDa): MES SDS buffer
Medium to large-size proteins (14–260 kDa): MOPs SDS buffer
Reduced samples: Antioxidant
Bis-tris transfer buffer
Bolt Bis-Tris Plus SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Bis-trisDenaturing: LDS sample buffer
Reducing: Sample reducing agent
Small to medium-sized proteins (6–260 kDa): MES SDS buffer
Medium to large-size proteins (14–260 kDa): MOPs SDS buffer
Reduced samples: Antioxidant
Bis-tris transfer buffer
Why LDS sample buffers
Bolt and NuPAGE LDS Sample Buffers are formulated with Coomassie G250 and phenol red as tracking dyes instead of bromophenol blue. Coomassie (SERVA) G250 gives a sharp dye front with both MES and MOPS SDS running buffers and migrates much closer to the moving ion front than bromophenol blue. Bromophenol blue runs more slowly than some peptides with the MES SDS Running Buffer. This ensures that small peptides do not run off the gel. The concentration of the tracking dye (Coomassie G250) is increased in Bolt and NuPAGE LDS Sample Buffers to enhance viewing of the dye front. LDS is used instead of SDS to allow the formulation of a 4X solution. The 4X solution provides the ability to have higher protein concentrations and less dilution of your protein sample.
Tris-glycine buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Tris-glycineDenaturing: Tris-glycine SDS sample buffer
Native: Tris-glycine native sample buffer
Denaturing: Tris-glycine SDS buffer
Native: Tris-glycine native buffer
Tris-glycine transfer buffer
Tris-acetate buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
High MW
(40-500 kDa)
Tris-acetateDenaturing: LDS sample buffer
Native: Tris-glycine native sample buffer
Denaturing: Tris-acetate SDS buffer
Native: Tris-glycine native buffer
Bis-tris transfer buffer
Tricine SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Low MW
(2.5-40 kDa)
TricineDenaturing: Tricine SDS sample bufferTricine SDS bufferTris-glycine transfer buffer
IEF gel recommended buffers
Zymography gel buffers & reagents
NativePAGE Bis-Tris gel buffers & reagents
Gel casting
When pouring your own polyacrylamide gels, choosing high-quality reagents can help you achieve optimal electrophoresis results. SureCast buffers and reagents make it quick and easy to make high quality handcast Tris-glycine gels.
SDS-PAGE gel recipe for pour-your-own Tris-glycine gels
Prepare resolving gel solution according to the volumes in the following table. The volumes provided in the table are for a single gel. Scale volumes proportionally based on the number of gels to be cast.
 Polyacrylamide %
Solution4%6%8%10%12%14%16%18%20%
SureCast Acrylamide (40%)0.8mL1.2mL1.6mL2.0mL2.4mL2.8mL3.3mL3.6mL4.0mL
SureCast Resolving Buffer2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL
Distilled water5.1mL4.7mL4.3mL3.9mL3.5mL3.1mL2.7mL2.3mL1.9mL
10% SureCast Ammonium Persulfate (APS)80µL80µL80µL80µL80µL80µL80µL80µL80µL
SureCast TEMED**8µL8µL8µL8µL8µL8µL8µL8µL8µL
**Add this last and mix well just before the gel is to be poured
Prepare stacking gel solution according to the following table. The volumes provided in the table are for a single gel. Scale volumes proportionally based on the number of gels to be cast. Note: Solutions do not require degassing.
***Add this last and mix well just before the gel is to be poured
Learn more about gel casting and find additional SDS-PAGE gel recipes.
Ordering quick links
NuPAGE Bis-Tris Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
NP0005 NuPAGE™ Antioxidant Each
54,75
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
21,50
NP0002 NuPAGE™ MES SDS Running Buffer (20X), 500 mL Each
102,65
Offre exceptionnelle en ligne
127,00
Économisez 24,35 (19%)
NP0001 NuPAGE™ MOPS SDS Running Buffer (20X), 500 mL Each
105,65
Offre exceptionnelle en ligne
128,00
Économisez 22,35 (17%)
NP000102 NuPAGE™ MOPS SDS Running Buffer (20X), 5 L Each
779,65
Offre exceptionnelle en ligne
942,00
Économisez 162,35 (17%)
NP000202 NuPAGE™ MES SDS Running Buffer (20X), 5 L Each
769,65
Offre exceptionnelle en ligne
932,00
Économisez 162,35 (17%)
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
192,00
NP0060 NuPAGE™ MES SDS Buffer Kit (for Bis-Tris Gels) Each
156,65
Offre exceptionnelle en ligne
190,00
Économisez 33,35 (18%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
235,00
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
233,65
Offre exceptionnelle en ligne
288,00
Économisez 54,35 (19%)
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
28,16
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Bolt Bis-Tris Plus Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
B0007 4X Bolt™ LDS Sample Buffer, 10 mL Each
18,65
Offre exceptionnelle en ligne
21,11
Économisez 2,46 (12%)
BT0005 Bolt™ Antioxidant Each
54,75
B0008 4X Bolt™ LDS Sample Buffer, 250 mL Each
144,65
Offre exceptionnelle en ligne
169,00
Économisez 24,35 (14%)
B0009 10X Bolt™ Sample Reducing Agent, 10 mL Each
68,65
Offre exceptionnelle en ligne
77,25
Économisez 8,60 (11%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
258,65
Offre exceptionnelle en ligne
283,00
Économisez 24,35 (9%)
B000202 20X Bolt™ MES SDS Running Buffer, 5 L Each
359,65
Offre exceptionnelle en ligne
399,00
Économisez 39,35 (10%)
B0001 20X Bolt™ MOPS SDS Running Buffer, 500 mL Each
80,00
B000102 20X Bolt™ MOPS SDS Running Buffer, 5 L Each
397,00
B0002 20X Bolt™ MES SDS Running Buffer, 500 mL Each
70,65
Offre exceptionnelle en ligne
78,50
Économisez 7,85 (10%)
B0004 10X Bolt™ Sample Reducing Agent, 250 μL Each
16,65
Offre exceptionnelle en ligne
18,06
Économisez 1,41 (8%)
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
42,65
Offre exceptionnelle en ligne
46,38
Économisez 3,73 (8%)
Tris-Glycine Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
LC3675 Tris-Glycine Transfer Buffer (25X) Each
68,65
Offre exceptionnelle en ligne
84,25
Économisez 15,60 (19%)
LC2675 Novex™ Tris-Glycine SDS Running Buffer (10X), 500 mL Each
46,65
Offre exceptionnelle en ligne
57,75
Économisez 11,10 (19%)
LC2676 Novex™ Tris-Glycine SDS Sample Buffer (2X) Each
40,88
LC26755 Novex™ Tris-Glycine SDS Running Buffer (10X), 5 L Each
267,65
Offre exceptionnelle en ligne
311,00
Économisez 43,35 (14%)
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
52,65
Offre exceptionnelle en ligne
63,25
Économisez 10,60 (17%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
39,63
Tris-Acetate Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
NP0005 NuPAGE™ Antioxidant Each
54,75
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
21,50
LA0041 NuPAGE™ Tris-Acetate SDS Running Buffer (20X) Each
99,65
Offre exceptionnelle en ligne
122,00
Économisez 22,35 (18%)
LA0050 NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007 Each
191,00
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
116,00
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
235,00
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
28,16
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
52,65
Offre exceptionnelle en ligne
63,25
Économisez 10,60 (17%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
39,63
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Tricine System Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
LC3675 Tris-Glycine Transfer Buffer (25X) Each
68,65
Offre exceptionnelle en ligne
84,25
Économisez 15,60 (19%)
LC1677 Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 Each
253,00
LC1676 Novex™ Tricine SDS Sample Buffer (2X) Each
45,96
LC1675 Novex™ Tricine SDS Running Buffer (10X) Each
168,65
Offre exceptionnelle en ligne
196,00
Économisez 27,35 (14%)
IEF Buffers
Zymogram Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
LC2675 Novex™ Tris-Glycine SDS Running Buffer (10X), 500 mL Each
46,65
Offre exceptionnelle en ligne
57,75
Économisez 11,10 (19%)
LC2676 Novex™ Tris-Glycine SDS Sample Buffer (2X) Each
40,88
LC26754 Novex™ Tris-Glycine SDS Running Buffer (10X), 4 x 1 L Each
246,65
Offre exceptionnelle en ligne
283,00
Économisez 36,35 (13%)
LC2670 Novex™ Zymogram Renaturing Buffer (10X) Each
90,00
LC2671 Novex™ Zymogram Developing Buffer (10X) Each
101,00
NativePAGE Bis-Tris Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
BN2008 NativePAGE™ Sample Prep Kit Each
362,00
BN2007 NativePAGE™ Running Buffer Kit Each
310,65
Offre exceptionnelle en ligne
327,00
Économisez 16,35 (5%)
BN2001 NativePAGE™ Running Buffer (20X) Each
286,65
Offre exceptionnelle en ligne
329,00
Économisez 42,35 (13%)
BN2004 NativePAGE™ 5% G-250 Sample Additive Each
99,25
BN2002 NativePAGE™ Cathode Buffer Additive (20X) Each
61,00
BN2003 NativePAGE™ Sample Buffer (4X) Each
99,75
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Handcast Buffers & Reagents
SDS sample buffers or loading buffers
Protein samples prepared for SDS-PAGE analysis are denatured by heating in the presence of a sample buffer containing 1–2% SDS or LDS with or without a reducing agent such as 20 mM DTT, 2-mercaptoethanol (BME) or TCEP. Sample buffers or loading buffers also contain glycerol so that they are heavier than water and sink neatly to the bottom of the buffer-submerged well when added to a gel. In addition, a negatively charged, low-molecular weight dye is also included in the sample buffer that will migrate at the buffer-front, enabling one to monitor the progress of electrophoresis. The most common tracking dyes for sample loading buffers are bromophenol blue, phenol red, and Coomassie blue. The table below summaries common sample buffers used in the different gel buffer systems.
Gel systemDenaturing sample buffer (final concentrations)Sample preparation
Tris-glycineTris-glycine SDS sample buffer: Tris HCl (63 mM), glycerol (10%), SDS (2%), bromophenol blue (0.0025%), pH 6.8Heat samples at 85°C for 2–5 minutes with sample buffer
Bis-TrisLDS sample buffer: Tris base (141 mM), Tris HCl (106 mM), LDS (2%), EDTA (0.51 mM), SERVA Blue G-250 (0.22 mM), phenol red (0.175 mM), pH 8.5Heat samples at 70°C for 10 minutes with sample buffer
Tris-AcetateLDS sample buffer: Tris base (141 mM), Tris HCl (106 mM), LDS (2%), EDTA (0.51 mM), SERVA Blue G-250 (0.22 mM), phenol red (0.175 mM), pH 8.5Heat samples at 70°C for 10 minutes with sample buffer
Tris-TricineTricine SDS sample buffer: Tris HCl (450 mM), glycerol (12%), SDS (4%), Coomassie Blue G (0.00075%), phenol red (0.0025%), pH 8.45Heat samples at 85°C for 2–5 minutes with sample buffer
ZymogramTris-glycine SDS: Tris HCl (63 mM), glycerol (10%), SDS (2%), bromophenol blue (0.0025%), pH 6.8Do not heat samples, load immediately onto gel after adding to sample
For fluorescence applications, we recommend that you use sample buffers that do not contain dyes that interfere in fluorescent channels (e.g., bromophenol blue). Fluorescent Compatible Sample Buffer does not contain bromophenol blue or other interfering dyes and can be used with reducing or nonreducing SDS-PAGE applications.
Find SDS sample buffer recipes in gel-specific product manuals in the documents tab.
Native sample buffers
Protein samples prepared for native PAGE applications are prepared in sample buffers or loading buffers that do not contain detergents to maintain the native state of the proteins. The loading buffers contain glycerol so that they are heavier than water and sink neatly to the bottom of the buffer-submerged well when added to a gel. In addition, a negatively charged, low-molecular weight dye is also included in the sample buffer that will migrate at the buffer-front, enabling one to monitor the progress of electrophoresis.
When preparing samples for native applications, protein samples should not be heated and loaded immediately onto the gel after mixing with the sample buffer.
Gel systemNative sample buffer (final concentrations)
Tris-glycineNative sample buffer: Tris HCl (100 mM), glycerol (10%), bromophenol blue (0.00025%), pH 8.6
Tris-acetateNative sample buffer: Tris HCl (100 mM), glycerol (10%), bromophenol blue (0.00025%), pH 8.6
IEFIEF sample buffer pH 3-7: Lysine (40 mM), glycerol (15%)
IEF sample buffer pH 3-10: Arginine (20 mM), Lysine (20 mM), glycerol (15%)
NativePAGE Bis-TrisNativePAGE Sample Buffer (4X): Bis-Tris (50 mM), 6 N HCl, NaCl (50 mM), Glycerol (10%), Ponceau S (0.001%), pH 7.2
Find native sample buffer recipes in gel-specific product manuals in the documents tab.
SDS running buffers
In protein electrophoresis (discontinuous buffer system), the primary anion in the gel is different (or discontinuous) from the primary anion in the running buffer. The tables below have the recommended SDS running buffers or native running buffers for each of the different gel chemistry buffering systems.
Gel systemSDS running buffer
Tris-glycineTris-glycine SDS: Tris base (25 mM), glycine (192 mM), SDS (0.1%), pH 8.3
Bis-TrisMES SDS: MES (50 mM), Tris base (50 mM), SDS (0.1%), EDTA (1 mM), pH 7.3
MOPS SDS: MOPS (50 mM), Tris base (50 mM), SDS (0.1%), EDTA (1 mM), pH 7.7
Tris-acetateTris-acetate SDS: Tris base (50 mM), Tricine (50 mM), SDS (0.1%), pH 8.24
Tris-tricineTricine-SDS: Tris base (100 mM), tricine (100 mM), SDS (0.1%), pH 8.3
Native running buffers
Gel systemNative running buffer
Tris-glycineTris-glycine native buffer: Tris base (25 mM), glycine (192 mM), pH 8.3
Tris-acetateTris-glycine native buffer: Tris base (25 mM), glycine (192 mM), pH 8.3
IEFIEF cathode buffer pH 3-7: Lysine (40 mM)
IEF cathode buffer pH 3-10: Arginine (20 mM), lysine (20 mM)
IEF anode buffer: phosphoric acid 85% (7 mM)
ZymogramTris-glycine SDS: Tris base (25 mM), glycine (192 mM), SDS (0.1%), pH 8.3
Find SDS running buffer recipes and native running buffer recipes in gel specific product manuals in the documents tab.
NuPAGE Bis-Tris SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Bis-trisDenaturing: LDS sample buffer
Reducing: Sample reducing agent
Small to medium-sized proteins (6–260 kDa): MES SDS buffer
Medium to large-size proteins (14–260 kDa): MOPs SDS buffer
Reduced samples: Antioxidant
Bis-tris transfer buffer
Bolt Bis-Tris Plus SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Bis-trisDenaturing: LDS sample buffer
Reducing: Sample reducing agent
Small to medium-sized proteins (6–260 kDa): MES SDS buffer
Medium to large-size proteins (14–260 kDa): MOPs SDS buffer
Reduced samples: Antioxidant
Bis-tris transfer buffer
Why LDS sample buffers
Bolt and NuPAGE LDS Sample Buffers are formulated with Coomassie G250 and phenol red as tracking dyes instead of bromophenol blue. Coomassie (SERVA) G250 gives a sharp dye front with both MES and MOPS SDS running buffers and migrates much closer to the moving ion front than bromophenol blue. Bromophenol blue runs more slowly than some peptides with the MES SDS Running Buffer. This ensures that small peptides do not run off the gel. The concentration of the tracking dye (Coomassie G250) is increased in Bolt and NuPAGE LDS Sample Buffers to enhance viewing of the dye front. LDS is used instead of SDS to allow the formulation of a 4X solution. The 4X solution provides the ability to have higher protein concentrations and less dilution of your protein sample.
Tris-glycine buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Broad-range MW (6-400 kDa)Tris-glycineDenaturing: Tris-glycine SDS sample buffer
Native: Tris-glycine native sample buffer
Denaturing: Tris-glycine SDS buffer
Native: Tris-glycine native buffer
Tris-glycine transfer buffer
Tris-acetate buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
High MW
(40-500 kDa)
Tris-acetateDenaturing: LDS sample buffer
Native: Tris-glycine native sample buffer
Denaturing: Tris-acetate SDS buffer
Native: Tris-glycine native buffer
Bis-tris transfer buffer
Tricine SDS buffers & reagents
Protein sampleGel chemistrySample buffersRunning bufferTransfer buffer
Low MW
(2.5-40 kDa)
TricineDenaturing: Tricine SDS sample bufferTricine SDS bufferTris-glycine transfer buffer
IEF gel recommended buffers
Zymography gel buffers & reagents
NativePAGE Bis-Tris gel buffers & reagents
Gel casting
When pouring your own polyacrylamide gels, choosing high-quality reagents can help you achieve optimal electrophoresis results. SureCast buffers and reagents make it quick and easy to make high quality handcast Tris-glycine gels.
SDS-PAGE gel recipe for pour-your-own Tris-glycine gels
Prepare resolving gel solution according to the volumes in the following table. The volumes provided in the table are for a single gel. Scale volumes proportionally based on the number of gels to be cast.
 Polyacrylamide %
Solution4%6%8%10%12%14%16%18%20%
SureCast Acrylamide (40%)0.8mL1.2mL1.6mL2.0mL2.4mL2.8mL3.3mL3.6mL4.0mL
SureCast Resolving Buffer2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL2.0mL
Distilled water5.1mL4.7mL4.3mL3.9mL3.5mL3.1mL2.7mL2.3mL1.9mL
10% SureCast Ammonium Persulfate (APS)80µL80µL80µL80µL80µL80µL80µL80µL80µL
SureCast TEMED**8µL8µL8µL8µL8µL8µL8µL8µL8µL
**Add this last and mix well just before the gel is to be poured
Prepare stacking gel solution according to the following table. The volumes provided in the table are for a single gel. Scale volumes proportionally based on the number of gels to be cast. Note: Solutions do not require degassing.
***Add this last and mix well just before the gel is to be poured
Learn more about gel casting and find additional SDS-PAGE gel recipes.
Ordering quick links
NuPAGE Bis-Tris Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
NP0005 NuPAGE™ Antioxidant Each
54,75
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
21,50
NP0002 NuPAGE™ MES SDS Running Buffer (20X), 500 mL Each
102,65
Offre exceptionnelle en ligne
127,00
Économisez 24,35 (19%)
NP0001 NuPAGE™ MOPS SDS Running Buffer (20X), 500 mL Each
105,65
Offre exceptionnelle en ligne
128,00
Économisez 22,35 (17%)
NP000102 NuPAGE™ MOPS SDS Running Buffer (20X), 5 L Each
779,65
Offre exceptionnelle en ligne
942,00
Économisez 162,35 (17%)
NP000202 NuPAGE™ MES SDS Running Buffer (20X), 5 L Each
769,65
Offre exceptionnelle en ligne
932,00
Économisez 162,35 (17%)
NP0050 NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels) Each
192,00
NP0060 NuPAGE™ MES SDS Buffer Kit (for Bis-Tris Gels) Each
156,65
Offre exceptionnelle en ligne
190,00
Économisez 33,35 (18%)
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
235,00
NP00061 NuPAGE™ Transfer Buffer (20X), 1L Each
233,65
Offre exceptionnelle en ligne
288,00
Économisez 54,35 (19%)
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
28,16
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Bolt Bis-Tris Plus Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
B0007 4X Bolt™ LDS Sample Buffer, 10 mL Each
18,65
Offre exceptionnelle en ligne
21,11
Économisez 2,46 (12%)
BT0005 Bolt™ Antioxidant Each
54,75
B0008 4X Bolt™ LDS Sample Buffer, 250 mL Each
144,65
Offre exceptionnelle en ligne
169,00
Économisez 24,35 (14%)
B0009 10X Bolt™ Sample Reducing Agent, 10 mL Each
68,65
Offre exceptionnelle en ligne
77,25
Économisez 8,60 (11%)
BT00061 Bolt™ Transfer Buffer (20X), 1L Each
258,65
Offre exceptionnelle en ligne
283,00
Économisez 24,35 (9%)
B000202 20X Bolt™ MES SDS Running Buffer, 5 L Each
359,65
Offre exceptionnelle en ligne
399,00
Économisez 39,35 (10%)
B0001 20X Bolt™ MOPS SDS Running Buffer, 500 mL Each
80,00
B000102 20X Bolt™ MOPS SDS Running Buffer, 5 L Each
397,00
B0002 20X Bolt™ MES SDS Running Buffer, 500 mL Each
70,65
Offre exceptionnelle en ligne
78,50
Économisez 7,85 (10%)
B0004 10X Bolt™ Sample Reducing Agent, 250 μL Each
16,65
Offre exceptionnelle en ligne
18,06
Économisez 1,41 (8%)
BT0006 Bolt™ Transfer Buffer (20X), 125mL Each
42,65
Offre exceptionnelle en ligne
46,38
Économisez 3,73 (8%)
Tris-Glycine Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
LC3675 Tris-Glycine Transfer Buffer (25X) Each
68,65
Offre exceptionnelle en ligne
84,25
Économisez 15,60 (19%)
LC2675 Novex™ Tris-Glycine SDS Running Buffer (10X), 500 mL Each
46,65
Offre exceptionnelle en ligne
57,75
Économisez 11,10 (19%)
LC2676 Novex™ Tris-Glycine SDS Sample Buffer (2X) Each
40,88
LC26755 Novex™ Tris-Glycine SDS Running Buffer (10X), 5 L Each
267,65
Offre exceptionnelle en ligne
311,00
Économisez 43,35 (14%)
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
52,65
Offre exceptionnelle en ligne
63,25
Économisez 10,60 (17%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
39,63
Tris-Acetate Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
NP0005 NuPAGE™ Antioxidant Each
54,75
NP0004 NuPAGE™ Sample Reducing Agent (10X), 250μL Each
21,50
LA0041 NuPAGE™ Tris-Acetate SDS Running Buffer (20X) Each
99,65
Offre exceptionnelle en ligne
122,00
Économisez 22,35 (18%)
LA0050 NuPAGE™ Tris-Acetate SDS Buffer Kit (for Tris-Acetate Gels), Contains 1 ea. LA0041, NP0004, NP0005, NP0007 Each
191,00
NP0009 NuPAGE™ Sample Reducing Agent (10X), 10mL Each
116,00
NP0008 NuPAGE™ LDS Sample Buffer (4X), 250mL Each
235,00
NP0007 NuPAGE™ LDS Sample Buffer (4X), 10mL Each
28,16
LC2672 Novex™ Tris-Glycine Native Running Buffer (10X) Each
52,65
Offre exceptionnelle en ligne
63,25
Économisez 10,60 (17%)
LC2673 Novex™ Tris-Glycine Native Sample Buffer (2X) Each
39,63
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Tricine System Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
LC3675 Tris-Glycine Transfer Buffer (25X) Each
68,65
Offre exceptionnelle en ligne
84,25
Économisez 15,60 (19%)
LC1677 Novex™ Tricine SDS Buffer Kit, includes LC1676 & LC1675 Each
253,00
LC1676 Novex™ Tricine SDS Sample Buffer (2X) Each
45,96
LC1675 Novex™ Tricine SDS Running Buffer (10X) Each
168,65
Offre exceptionnelle en ligne
196,00
Économisez 27,35 (14%)
IEF Buffers
Zymogram Buffers
NativePAGE Bis-Tris Buffers
N° de catalogue Nom Taille Prix (EUR) Quantité
BN2008 NativePAGE™ Sample Prep Kit Each
362,00
BN2007 NativePAGE™ Running Buffer Kit Each
310,65
Offre exceptionnelle en ligne
327,00
Économisez 16,35 (5%)
BN2001 NativePAGE™ Running Buffer (20X) Each
286,65
Offre exceptionnelle en ligne
329,00
Économisez 42,35 (13%)
BN2004 NativePAGE™ 5% G-250 Sample Additive Each
99,25
BN2002 NativePAGE™ Cathode Buffer Additive (20X) Each
61,00
BN2003 NativePAGE™ Sample Buffer (4X) Each
99,75
NP0006 NuPAGE™ Transfer Buffer (20X), 125mL Each
37,65
Offre exceptionnelle en ligne
49,59
Économisez 11,94 (24%)
Handcast Buffers & Reagents
For Research Use Only. Not for use in diagnostic procedures.