Automated extraction of nucleic acids, cells, exosomes or proteins is made possible by combining the right magnetic bead-based kit with a KingFisher sample purification system. Available kits, individual reagents, and downloadable protocols enable sample preparation that is easy and efficient. Each sample type and application require different chemistries to effectively extract the target analyte. For example, blood, saliva, and buccal samples are concentrated sources of long DNA fragments, while cell-free DNA sample sources are high-volume, low-concentration of short fragments. It takes different beads and different binding and buffer chemistry to optimally extract DNA from these sources. Explore our selection of kits and reagents for your target isolate and research application below.
Kit, reagent, and protocol selection tool
Use our selection tool to find the right magnetic bead-based kit for your automated sample preparation. MagMAX, Dynabeads, and SMART Digest kits can purify nucleic acids, proteins, cells, exosomes, and virus from a wide range of sample types and are optimized for automated Kingfisher purification systems. 
Go directly to our Kit, Reagent, and Protocol Selection Tool
Downloading protocols
KingFisher BindIt or BindIx software is included free with all KingFisher instruments so that you can create, download, run, modify and store protocols for your applications. The software allows you to define and edit steps, parameters, plates, and reagents for every part of your protocol—bind, wash and elute.
For any MagMAX or Dynabeads kits or reagents in the Kits, Reagents, and Protocols Selection Tool, click View Protocols to review automated protocols for KingFisher instruments. Then follow these steps to download and install the protocols you want.
KingFisher BindIx software (for KingFisher Apex systems)
KingFisher BindIt software (for KingFisher Flex and Duo Prime systems)
  1. Make sure you have BindIt 4.1 Software installed on the computer you use with your KingFisher instrument. Download the latest BindIt Software here.
  2. Find the kit you want in the Kit, Reagent, and Protocol Selection Tool and click View Protocols. You’ll see the product page for that kit.
  3. Scroll to the Protocols section at the bottom and click the protocol you want to download. Make sure it’s for the correct instrument.
  4. Save the protocol on your computer in the preferred folder.
  5. Connect your PC to your KingFisher instrument with a USB cable and select Home > Connect in BindIt Software.
  6. You can start the protocol from the BindIt Software or transfer it onto the instrument. If you transfer the protocol, select Home > Transfer in BindIt Software.
  7. Select Upload and find the saved protocol. Click Open.
  8. Select the folder (category) in which you want to store the protocol on the instrument. Your protocol can be now found in the instrument memory.
  9. Disconnect your instrument from BindIt and check using the keypad that your protocol is in the desired folder.
Liquid biopsy and cancer research
Liquid biopsy applications are at the forefront of modern cancer research with tissue biopsies remaining widely used for clinical cancer diagnosis. Combinatorial testing approaches provide researchers insight into the presence of cancer, tumor progression, and treatment response. This testing begins with the sample. Whether it is isolating circulating tumor DNA from whole blood or extracting nucleic acids from FFPE tissue, KingFisher instruments and kits can support with comprehensive solutions that help provide the analyte purity and yield needed to obtain accurate and reliable results. 
DNA & RNA isolation from FFPE tissue for cancer research
Clinical cancer research samples are routinely formalin fixed and paraffin embedded (FFPE) for histopathological analysis and tissue archiving. This method preserves the tissue and allows for long-term storage, but the extensive chemical crosslinking of nucleic acids can complicate downstream processing for applications such as next-generation sequencing (NGS).
With the MagMAX FFPE DNA/RNA Ultra Kit and KingFisher instruments, you can sequentially isolate DNA, RNA, microRNA, or both DNA and RNA from FFPE and fixed samples. The genomic DNA (gDNA) extracted is high quality with yields comparable to fresh-frozen blood. The kit is exceptional for use with Ion Torrent™ Oncomine™ assays as well as PCR and NGS, even when the FFPE tissue is more than 25 years old.
Sample FFPE isolation workflow
Sample FFPE Isolation workflow
Processing multiple FFPE samples?
Automated purification of cfDNA and cfTNA from liquid biopsy samples
“Liquid biopsies” of cell-free DNA (cfDNA) or total nucleic acids (cfTNA) from plasma, serum, or urine represent a non-invasive approach to detect and monitor targets associated with disease states such as cancer. MagMAX cell-free nucleic acid kits on KingFisher instruments are exceptional for liquid biopsy research. They are optimized specifically for enrichment of cell-free DNA (cfDNA) and total nucleic acid (cfTNA), not genomic DNA (gDNA), which means increased recovery and lower starting volumes. 
Sample cfDNA or cfTNA liquid biopsy workflow
Sample cfDNA or cfTNA liquid biopsy workflow
Automated CTC isolation from whole blood
In the oncology field, circulating tumor cells (CTCs) are gaining importance as prognostic markers and as tools for research into the monitoring of treatment response. Due to of the low number of CTCs in circulation, highly sensitive methods are necessary to capture and detect down to single cells. Dynabeads magnetic beads provide an automation-friendly tool for isolation of these circulating biomarkers for downstream analyses.
CTC isolation and enrichment sample workflows
automated workflows for circulating tumor cell isolation
Exosome & extracellular vesicle isolation from liquid biopsies
A highly specific approach to exosome isolation is immunomagnetic separation using Dynabeads magnetic beads. Immunomagnetic separation is well suited for purifying exosome subpopulations from liquid biopsy samples. Exosome-specific antibodies are bound to the surface of these beads for a more efficient purification of exosomes.
Automated bead-based target isolation workflow
Exosome isolation workflow diagram
Exosome isolation workflow using an automated method.
Sequential DNA & RNA isolation from blood and bone marrow
Isolation of nucleic acids from hematological samples is crucial for understanding and identifying cancer-causing genetic alterations. The MagMAX Sequential DNA/RNA Kit offers an automated workflow solution to sequentially isolate both genomic DNA and total RNA in one protocol, which enables a more comprehensive downstream analysis.
Sequential DNA & RNA isolation workflow
sequential isolation of DNA and RNA for NGS workflow diagram
Build a custom cancer workflow to fit your research needs
Banner showing 3D rendered cancer cells
Automated gDNA isolation for genomics applications
Isolating genomic DNA (gDNA) from different sample types has historically required multiple purification kits. With the MagMAX DNA Multi-Sample Ultra 2.0 Kit or KingFisher Flex Ready DNA Ultra 2.0 Prefilled plates and KingFisher instruments, you can isolate gDNA from 50 µL to 2mL of whole blood, saliva, buffy coat, buccal swabs, or other biological samples. The resulting purified gDNA is excellent for many downstream molecular biology applications such as qPCR, NGS, and microarray analysis.
One easily accessible, non-invasive, reliable source of gDNA suitable for downstream genetic analysis is saliva. The MagMAX gDNA Saliva Kit, developed for a leading genomics company for use with KingFisher instruments, is specifically optimized for saliva gDNA isolation.
Sample gDNA workflow
Sample gDNA workflow
Sample gDNA from saliva workflow
Sample gDNA from saliva workflow
Automated RNA isolation for RNA-based therapeutics
Total RNA and miRNA
Isolating different types of RNA from different sample types for gene expression studies has historically required multiple purification kits. The MagMAX mirVana Total RNA Isolation Kit is optimized to extract Total RNA, including small RNAs and microRNAs (miRNAs), from most biological samples, including plasma/serum, whole blood, solid tissue, cultured cells, urine, and more. Extraction is phenol-free and non-organic.
Sample total RNA or miRNA workflow
Sample total RNA or miRNA workflow
mRNA
Research on transcriptomics requires messenger RNA (mRNA), which can be sourced from a wide variety of samples. Dynabeads mRNA isolation beads specifically capture mRNA molecules from a wide variety of crude samples by exploiting the base pairing between the polyA residues at the 3’ end of most mRNA and the Oligo(dT)25 residues covalently coupled to the surface of the Dynabeads. Automated protocols on KingFisher instruments take only 15 minutes and the isolated mRNA is suitable for use in all downstream applications.
Sample viral/pathogen analysis workflow
Sample mRNA research workflow
Nucleic acid isolation from microbes
Microbes encompass a wide range of organisms, including bacteria, archaea, fungi, viruses, and protozoa. Understanding the diversity and characteristics of these different types of microbes is essential for advancing various scientific disciplines and addressing global challenges. The isolation of nucleic acids from microbes is essential for studying their genetic makeup and understanding their functions and interactions. It enables researchers to uncover valuable information about microbial diversity, evolution, and ecological roles. KingFisher instruments and MagMAX kits enable exceptional pathogen lysis and recovery of nucleic acid whether it is from viral particles or the toughest of pathogens like gram-positive bacteria, yeast, and fungi.
Microbiome research
The human microbiome has emerged as a key player in the diagnosis and treatment of disease, but identifying and quantifying the microbes that make it up constitute a major challenge. Although metagenomics sequencing technology can providing a comprehensive profile of all the genes in an environmental or human sample, it requires an adequate sample of pure, high-quality DNA and RNA. The MagMAX Prime Viral/Pathogen NA Isolation Kit or MagMAX Microbiome Ultra Nucleic Acid Isolation Kit with KingFisher instruments can provide an efficient automated nucleic acid isolation workflow for microbiome research applications.
Sample microbiome research workflow
decorative image
Wastewater surveillance
Wastewater and sewage surveillance is a useful tool for monitoring the regional spread of viral and bacterial pathogens. Surveillance at different levels of operation, such as a university dormitory or an entire city, has been shown to be a reliable way to detect the viruses before it becomes widespread in a population. Wastewater surveillance can be used to monitor other viral targets such as influenza and norovirus. The MagMAX Wastewater Kits offer an efficient and simple method for extracting high-quality nucleic acids from wastewater, sewage, or sludge samples for disease surveillance workflows. The purified DNA and RNA is excellent for use in a variety of downstream applications such as quantitative real-time PCR, digital PCR, or next-generation sequencing.
Sample wastewater research workflow
Sample wastewater research workflow
Nucleic acid purification for pathogen detection
The detection of viruses in human biofluids is crucial for infectious disease research and management. Viral load testing is used to assess the efficacy of antiviral therapy, as well as to monitor disease progression and uncover the presence of an infection at an earlier stage. MagMAX viral/pathogen kits on KingFisher instruments provide a sensitive and simple method for nucleic acid extraction from samples containing viruses or other pathogens.
Note: For specific information about using MagMAX Viral/Pathogen kits for SARS-CoV-2 testing, see SARS-CoV-2 testing above.
Sample viral/pathogen analysis workflow
decorative image
SARS-CoV-2 testing
KingFisher systems are on the front line of the battle against SARS-CoV-2 by helping researchers accelerate their RNA extraction with maximized throughput to help handle the huge demand for fast and accurate testing. MagMAX viral/pathogen kits on KingFisher instruments provide a sensitive and easy method for nucleic acid extraction for SARS-CoV-2 research applications, and protocols have recently been streamlined to further increase throughput.
Automated isolation of DNA and RNA for plant genomics
Extracting DNA and RNA from plants brings unique challenges in dealing with carbohydrates, phenolics, and other compounds abundant in plant tissues. MagMAX™ Plant DNA Isolation Kit, automated with KingFisher instruments, can isolate nucleic acids from a wide variety of plant species and tissue types.
Sample plant DNA workflow
decorative image
Sample plant RNA workflow
Sample plant RNA workflow
Immunoprecipitation and digestion for protein analysis
Immunoprecipitation
Immunoprecipitation (IP) or co-immunoprecipitation (co-IP) can be used to isolate and concentrate a particular protein or protein complex from a sample containing many thousands of different proteins. Dynabeads magnetic beads, which helps maximize yield and reproducibility and minimize nonspecific binding, have become trusted for magnetic bead-based IP. With automation protocols now available for KingFisher instruments, you can be ready to detect and analyze proteins in as little as 40 minutes with minimal hands-on time.
Sample immunoprecipitation (IP) workflow
Sample immunoprecipitation (IP) workflow
Recommended kits and products for immunoprecipitation
Peptide mapping and quantitation
Peptide mapping is a critical workflow in biotherapeutic protein characterization and is essential for elucidating the primary amino acid structure of proteins. In order to generate a peptide map, the therapeutic protein must first be digested into its constituent peptides via a chemical or enzymatic reaction. But methods currently employed for digestion and mapping involve lengthy, multi-step, manual processes subject to low levels of reproducibility, sensitivity, and data confidence.
Thermo Scientific SMART Digest magnetic kits paired with KingFisher instruments can shorten a 24-hour process to an hour or less, automating much of the hands-on work and eliminating sources of variation and manual error.
Sample peptide workflow
Sample Peptide mapping workflow
Sample peptide quantitation workflow
sample peptide quantitation workflow
Commercial supply
Our commercial supply team specializes in helping you bring your solutions to market faster. Connect with our team and learn how we can support your next innovative project with reagents and consumables, instruments, and custom kit package solutions.