GlutaMAX supplement product family photo
Choose Gibco L-glutamine for your cell culture
L-glutamine is an important amino acid supplement commonly added to mammalian cell culture media. L-glutamine supports the growth of cells in cell culture by serving as an auxiliary energy source, especially when the cells are rapidly dividing. L-glutamine is also important in the production of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, other amino acids, and plays a role in protein synthesis and glucose production.
Gibco L-glutamine is offered in both liquid and powdered forms. Powdered L-glutamine is supplied as a USP-grade chemical, and liquid formulations are supplied as ready-to-use 200 mM stock solutions. The optimal concentration of L-glutamine for cell culture is dependent upon the cell type and medium used, but generally falls in the range of 2–6 mM.
Catalog # Name Size Price (USD) Qty
21051024 L-Glutamine, 100 g Each
131.65
Online Exclusive
141.00
Save 9.35 (7%)
25030164 L-Glutamine (200 mM), 20 x 100 mL Each
667.65
Online Exclusive
726.00
Save 58.35 (8%)
21051040 L-Glutamine, 1 kg Each
773.65
Online Exclusive
832.00
Save 58.35 (7%)
25030081 L-Glutamine (200 mM), 100 mL Each
37.65
Online Exclusive
40.88
Save 3.23 (8%)
25030149 L-Glutamine (200 mM), 20 mL Each
13.65
Online Exclusive
14.67
Save 1.02 (7%)
cell icon
Enable improved cell viability with GlutaMAX supplement
Explore the differences between L-glutamine and GlutaMAX supplement with the function and performance data provided below. Learn how you can get the most from your cell cultures with GlutaMAX supplement.
L-glutamine vs GlutaMAX supplement
In aqueous solutions such as cell culture media, L-glutamine spontaneously degrades, generating toxic ammonia and pyrrolidine carboxylic acid as byproducts. The rate at which degradation of L-glutamine proceeds is a function of time, temperature, and pH. This L-glutamine degradation can be exacerbated as pH levels rise in the presence of bases like phosphate or bicarbonate. Likewise, L-glutamine degradation can occur at acidic pH levels. To minimize the toxic effects of L-glutamine degradation on your cells, you can adopt a fed-batch strategy to continuously feed low levels of L-glutamine into the culture.
In contrast to standard L-glutamine, which is more sensitive to environmental factors such as pH, Gibco GlutaMAX supplement is a dipeptide, L-alanyl-L-glutamine, which is more stable in aqueous solutions and does not spontaneously degrade. Instead, cells gradually release aminopeptidases that hydrolyze the dipeptide, slowly releasing L-alanine and L-glutamine into the culture media (Figure 2). The L-glutamine and L-alanine can then be taken up by the cells and utilized for protein production or in the TCA cycle. 
pyrrolidone-carboxylic-acid
Figure 1. L-glutamine spontaneously decomposes into ammonia and pyrrolidone carboxylic acid at a rate dependent on pH and temperature.
controlled-delivery-glutamine
Figure 2. GlutaMAX supplement allows for controlled delivery of L-glutamine from media to cells in culture.
stability-of-gmax
Figure 3. L-glutamine degrades faster than GlutaMAX Supplement in media at 37°C. DMEM was supplemented with GlutaMAX Supplement or L-glutamine, dispensed into vials, and stored at 37°C. Samples were taken daily and frozen at –20°C. Levels of GlutaMAX Supplement and L-glutamine were determined by HPLC.
ammonia-levels
Figure 4. Ammonia levels in supplemented media. DMEM was supplemented with GlutaMAX Supplement or L-glutamine, dispensed into vials, and stored at 37°C. Samples were taken daily and frozen at –20°C. Levels of ammonia were determined by HPLC.
Using GlutaMAX supplement in cell culture
Gibco GlutaMAX supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in your cell culture media, resulting in efficient energy metabolism and high-growth yield, without the detrimental effects of excess ammonia on your cells.
Microscopic view of fluorescently stained MSCs indicating CD105
Figure 5. Human mesenchymal stem cells cultured with GlutaMAX media. They are stained with DAPI nuclear stain and primary CD105 antibody bound to a secondary Invitrogen Alexa Fluor 488 antibody.
Microscopic view of fluorescently stained MSCs in adipogenic differentiating conditions
Figure 6. Human mesenchymal stem cells cultured with GlutaMAX media under adipogenic differentiating conditions. They are stained with Hoechst 33342 nuclear stain, Invitrogen LipidTOX Green neutral lipid, and Invitrogen MitoTracker Red.
How does GlutaMAX supplement improve your cell culture?
Using GlutaMAX supplement in cell culture in place of L-glutamine can improve cell viability and growth, potentially increasing productivity levels. Figures 7 and 8 show an AE-1 cell growth curve and recombinant IgG1production. The GlutaMAX supplement culture demonstrates improved cell viability and productivity.
In addition, GlutaMAX supplement can extend the life of your cell culture, thus reducing the number of times the cells must be passaged, saving you time and money. Figure 9 compares MDBK cells cultured in DMEM with 10% FBS and L-glutamine or GlutaMAX supplement. Cells cultured in GlutaMAX supplement reach peak density two days later and viability declines less rapidly than that observed in cultures with L-glutamine supplementation. The slight increase in lag phase is attributed to the time needed to release the peptidase and digest the dipeptide, allowing a gradual increase in availability of L-glutamine to the cells. (Appl Biochem Biotechnol 1991, 30:29.)
Bar graph of viable cells vs time, showing GlutaMAX supplement supported increased cell density compared to other media formulations
Figure 7. Cell growth. AE-1 mouse myeloma cells were seeded at 1x105 cells/mL. Samples were taken daily after three days and evaluated in triplicate for cell density. Cell viability was determined by trypan blue exclusion. Cell density continued to increase in samples containing GlutaMAX supplement past the point when L-glutamine containing samples decreased in cell density.
Bar graph of IgG1 vs time, showing GlutaMAX supplement supported increased IgG1 production compared to other media formulations
Figure 8. IgG1 production. The samples from Figure 7 were also examined by ELISA for IgG1 production. GlutaMAX supplement-containing samples outperformed L-glutamine-containing samples.
Line graph of cell count vs time, showing GlutaMAX supplement supported better growth over time than L-glutamine
Figure 9. Cell count. MDBK cells were seeded at approximately 1x105 cells/flask in D-MEM with 10% FBS and L-glutamine or GlutaMAX supplement in 25 cm2 T-flasks.
Description
Classical medium with L-glutamineClassical medium without L-glutamineQuantityMedium with GlutaMAX Supplement
 Cat. No.Cat. No.QuantityCat. No.Quantity
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
Low glucose, contains sodium pyruvate
11885076 1,000 mL10567014
(in EU 21885025)
500 mL
11885084 500 mL
11885092 10 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains sodium pyruvate
11995040 1,000 mL10569010
(in EU 31966021)
500 mL
11995081 6 x 1,000 mL
1199506510313021500 mL
11995073 10 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains no sodium pyruvate
11965084119600511,000 mL10566016
(in EU 61965026)
500 mL
11965126119600776 x 1,000 mL
1196509211960044500 mL
119651181196006910 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains HEPES buffer but no sodium pyruvate
12430047 1,000 mL10564011
(in EU 32430027)
500 mL
12430054 500 mL
12430062 10 x 500 mL
DMEM/F-12 (1X), liquid, 1:111320033 500 mL10565018
(in EU 31331028)
500 mL
Ham’s F-12 Nutrient Mix (1X), liquid11765047 1,000 mL31765035
(in EU 31765027)
500 mL
11765070 6 x 1,000 mL
11765054 500 mL
11765062 10 x 500 mL
Iscove’s Modified Dulbecco’s Medium (IMDM) (1X), liquid12440046 1,000 mL31980030
(in EU 31980022)
500 mL
12440053 500 mL
12440061 10 x 500 mL
Minimum Essential Medium (MEM) alpha (1X), liquid
Contains no ribonucleosides or deoxyribonucleosides
12561049 1,000 mL32561037
(in EU 32561029)
500 mL
12561056 500 mL
Minimum Essential Medium (MEM) alpha (1X), liquid
Contains ribonucleosides and deoxyribonucleosides
12571048 1,000 mL32571036
(in EU 32571028)
500 mL
12571063 500 mL
12571071 10 x 500 mL
Minimum Essential Medium (MEM), liquid
Contains Earle’s salts
11095072110900731,000 mL41090036
(in EU 41090028)
500 mL
1109508011090081500 mL
110950981109009910 x 500 mL
Minimum Essential Medium (MEM), liquid
Contains Earle’s salts and HEPES buffer
 12360038500 mL42360032
(in EU 42360024)
500 mL
Opti-MEM I Reduced-Serum Medium (1X), liquid31985062 100 mL51985034
(in EU 51985026)
500 mL
31985070 500 mL
31985088 1,000 mL
RPMI 1640 Medium (1X), liquid11875085218700841,000 mL61870036
(in EU 61870010)
500 mL
11875135 6 x 1,000 mL
1187509321870076500 mL
118751192187009210 x 500 mL
11875101 100 mL
11875127 20 x 100 mL
RPMI 1640 Medium (1X), liquid
Contains HEPES buffer
22400071 1,000 mL72400047
(in EU 72400021)
500 mL
22400089 500 mL
22400105 10 x 500 mL
    GlutaMAX Supplement
 Cat. No. QuantityCat. No.Quantity
L-Glutamine 200 mM (100X), liquid25030149 20 mL35050061
(in EU 35050038)
100 mL
25030081 100 mL
L-glutamine vs GlutaMAX supplement
In aqueous solutions such as cell culture media, L-glutamine spontaneously degrades, generating toxic ammonia and pyrrolidine carboxylic acid as byproducts. The rate at which degradation of L-glutamine proceeds is a function of time, temperature, and pH. This L-glutamine degradation can be exacerbated as pH levels rise in the presence of bases like phosphate or bicarbonate. Likewise, L-glutamine degradation can occur at acidic pH levels. To minimize the toxic effects of L-glutamine degradation on your cells, you can adopt a fed-batch strategy to continuously feed low levels of L-glutamine into the culture.
In contrast to standard L-glutamine, which is more sensitive to environmental factors such as pH, Gibco GlutaMAX supplement is a dipeptide, L-alanyl-L-glutamine, which is more stable in aqueous solutions and does not spontaneously degrade. Instead, cells gradually release aminopeptidases that hydrolyze the dipeptide, slowly releasing L-alanine and L-glutamine into the culture media (Figure 2). The L-glutamine and L-alanine can then be taken up by the cells and utilized for protein production or in the TCA cycle. 
pyrrolidone-carboxylic-acid
Figure 1. L-glutamine spontaneously decomposes into ammonia and pyrrolidone carboxylic acid at a rate dependent on pH and temperature.
controlled-delivery-glutamine
Figure 2. GlutaMAX supplement allows for controlled delivery of L-glutamine from media to cells in culture.
stability-of-gmax
Figure 3. L-glutamine degrades faster than GlutaMAX Supplement in media at 37°C. DMEM was supplemented with GlutaMAX Supplement or L-glutamine, dispensed into vials, and stored at 37°C. Samples were taken daily and frozen at –20°C. Levels of GlutaMAX Supplement and L-glutamine were determined by HPLC.
ammonia-levels
Figure 4. Ammonia levels in supplemented media. DMEM was supplemented with GlutaMAX Supplement or L-glutamine, dispensed into vials, and stored at 37°C. Samples were taken daily and frozen at –20°C. Levels of ammonia were determined by HPLC.
Using GlutaMAX supplement in cell culture
Gibco GlutaMAX supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in your cell culture media, resulting in efficient energy metabolism and high-growth yield, without the detrimental effects of excess ammonia on your cells.
Microscopic view of fluorescently stained MSCs indicating CD105
Figure 5. Human mesenchymal stem cells cultured with GlutaMAX media. They are stained with DAPI nuclear stain and primary CD105 antibody bound to a secondary Invitrogen Alexa Fluor 488 antibody.
Microscopic view of fluorescently stained MSCs in adipogenic differentiating conditions
Figure 6. Human mesenchymal stem cells cultured with GlutaMAX media under adipogenic differentiating conditions. They are stained with Hoechst 33342 nuclear stain, Invitrogen LipidTOX Green neutral lipid, and Invitrogen MitoTracker Red.
How does GlutaMAX supplement improve your cell culture?
Using GlutaMAX supplement in cell culture in place of L-glutamine can improve cell viability and growth, potentially increasing productivity levels. Figures 7 and 8 show an AE-1 cell growth curve and recombinant IgG1production. The GlutaMAX supplement culture demonstrates improved cell viability and productivity.
In addition, GlutaMAX supplement can extend the life of your cell culture, thus reducing the number of times the cells must be passaged, saving you time and money. Figure 9 compares MDBK cells cultured in DMEM with 10% FBS and L-glutamine or GlutaMAX supplement. Cells cultured in GlutaMAX supplement reach peak density two days later and viability declines less rapidly than that observed in cultures with L-glutamine supplementation. The slight increase in lag phase is attributed to the time needed to release the peptidase and digest the dipeptide, allowing a gradual increase in availability of L-glutamine to the cells. (Appl Biochem Biotechnol 1991, 30:29.)
Bar graph of viable cells vs time, showing GlutaMAX supplement supported increased cell density compared to other media formulations
Figure 7. Cell growth. AE-1 mouse myeloma cells were seeded at 1x105 cells/mL. Samples were taken daily after three days and evaluated in triplicate for cell density. Cell viability was determined by trypan blue exclusion. Cell density continued to increase in samples containing GlutaMAX supplement past the point when L-glutamine containing samples decreased in cell density.
Bar graph of IgG1 vs time, showing GlutaMAX supplement supported increased IgG1 production compared to other media formulations
Figure 8. IgG1 production. The samples from Figure 7 were also examined by ELISA for IgG1 production. GlutaMAX supplement-containing samples outperformed L-glutamine-containing samples.
Line graph of cell count vs time, showing GlutaMAX supplement supported better growth over time than L-glutamine
Figure 9. Cell count. MDBK cells were seeded at approximately 1x105 cells/flask in D-MEM with 10% FBS and L-glutamine or GlutaMAX supplement in 25 cm2 T-flasks.
Description
Classical medium with L-glutamineClassical medium without L-glutamineQuantityMedium with GlutaMAX Supplement
 Cat. No.Cat. No.QuantityCat. No.Quantity
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
Low glucose, contains sodium pyruvate
11885076 1,000 mL10567014
(in EU 21885025)
500 mL
11885084 500 mL
11885092 10 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains sodium pyruvate
11995040 1,000 mL10569010
(in EU 31966021)
500 mL
11995081 6 x 1,000 mL
1199506510313021500 mL
11995073 10 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains no sodium pyruvate
11965084119600511,000 mL10566016
(in EU 61965026)
500 mL
11965126119600776 x 1,000 mL
1196509211960044500 mL
119651181196006910 x 500 mL
Dulbecco’s Modified Eagle Medium (DMEM) (1X), liquid
High glucose, contains HEPES buffer but no sodium pyruvate
12430047 1,000 mL10564011
(in EU 32430027)
500 mL
12430054 500 mL
12430062 10 x 500 mL
DMEM/F-12 (1X), liquid, 1:111320033 500 mL10565018
(in EU 31331028)
500 mL
Ham’s F-12 Nutrient Mix (1X), liquid11765047 1,000 mL31765035
(in EU 31765027)
500 mL
11765070 6 x 1,000 mL
11765054 500 mL
11765062 10 x 500 mL
Iscove’s Modified Dulbecco’s Medium (IMDM) (1X), liquid12440046 1,000 mL31980030
(in EU 31980022)
500 mL
12440053 500 mL
12440061 10 x 500 mL
Minimum Essential Medium (MEM) alpha (1X), liquid
Contains no ribonucleosides or deoxyribonucleosides
12561049 1,000 mL32561037
(in EU 32561029)
500 mL
12561056 500 mL
Minimum Essential Medium (MEM) alpha (1X), liquid
Contains ribonucleosides and deoxyribonucleosides
12571048 1,000 mL32571036
(in EU 32571028)
500 mL
12571063 500 mL
12571071 10 x 500 mL
Minimum Essential Medium (MEM), liquid
Contains Earle’s salts
11095072110900731,000 mL41090036
(in EU 41090028)
500 mL
1109508011090081500 mL
110950981109009910 x 500 mL
Minimum Essential Medium (MEM), liquid
Contains Earle’s salts and HEPES buffer
 12360038500 mL42360032
(in EU 42360024)
500 mL
Opti-MEM I Reduced-Serum Medium (1X), liquid31985062 100 mL51985034
(in EU 51985026)
500 mL
31985070 500 mL
31985088 1,000 mL
RPMI 1640 Medium (1X), liquid11875085218700841,000 mL61870036
(in EU 61870010)
500 mL
11875135 6 x 1,000 mL
1187509321870076500 mL
118751192187009210 x 500 mL
11875101 100 mL
11875127 20 x 100 mL
RPMI 1640 Medium (1X), liquid
Contains HEPES buffer
22400071 1,000 mL72400047
(in EU 72400021)
500 mL
22400089 500 mL
22400105 10 x 500 mL
    GlutaMAX Supplement
 Cat. No. QuantityCat. No.Quantity
L-Glutamine 200 mM (100X), liquid25030149 20 mL35050061
(in EU 35050038)
100 mL
25030081 100 mL
GlutaMAX supplement composition
We offer the GlutaMAX supplement as a standalone 200 mM solution of the L-alanyl-L-glutamine dipeptide, which can be used as a direct substitute for L-glutamine at equimolar concentrations in your current cell culture media formulation. GlutaMAX supplement is stable at room temperature, meaning you can avoid freeze-thaw cycles and decrease contamination risk from frequent liquid-handling steps.
Product image of Gibco GlutaMAX Supplement
Taking your work to the clinic?  Be sure to check out CTS GlutaMAX supplement
Protect your cell cultures with Gibco Penicillin-Streptomycin-GlutaMAX supplement
Catalog # Name Size Price (USD) Qty
35050061 GlutaMAX™ Supplement, 100 mL Each
55.50
GlutaMAX media formulations
In addition to the standalone supplement, you may select from many media formulations in which the GlutaMAX dipeptide is substituted for L-glutamine. These options include DMEM, MEM, RPMI, Opti-MEM and other basal media. The Gibco GlutaMAX supplement and media are suitable for both adherent and suspension mammalian cell cultures, with little to no further adaptation required.
Image showing Gibco bottle with red medium
Catalog # Name Size Price (USD) Qty
10564029 DMEM, high glucose, GlutaMAX™ Supplement, HEPES, 10 x 500 mL Each
562.65
Online Exclusive
622.00
Save 59.35 (10%)
10567022 DMEM, low glucose, GlutaMAX™ Supplement, pyruvate, 10 x 500 mL Each
445.00
A4192101 BenchStable™ DMEM, 500 mL Each
44.65
Online Exclusive
48.37
Save 3.72 (8%)
10567014 DMEM, low glucose, GlutaMAX™ Supplement, pyruvate, 500 mL Each
50.50
A4192102 BenchStable™ DMEM, 10 x 500 mL Each
399.65
Online Exclusive
431.00
Save 31.35 (7%)
10566016 DMEM, high glucose, GlutaMAX™ Supplement, 500 mL Each
43.65
Online Exclusive
48.29
Save 4.64 (10%)
10564011 DMEM, high glucose, GlutaMAX™ Supplement, HEPES, 500 mL Each
65.65
Online Exclusive
72.75
Save 7.10 (10%)
10564045 DMEM, high glucose, GlutaMAX™ Supplement, HEPES, 10 L Each
1,250.65
Online Exclusive
1,382.00
Save 131.35 (10%)
10566024 DMEM, high glucose, GlutaMAX™ Supplement, 10 x 500 mL Each
388.65
Online Exclusive
429.00
Save 40.35 (9%)
10566032 DMEM, high glucose, GlutaMAX™ Supplement, 5 L Each
638.65
Online Exclusive
704.00
Save 65.35 (9%)
10569044 DMEM, high glucose, GlutaMAX™ Supplement, pyruvate, 10 x 500 mL Each
392.65
Online Exclusive
423.00
Save 30.35 (7%)
10569010 DMEM, high glucose, GlutaMAX™ Supplement, pyruvate, 500 mL Each
44.65
Online Exclusive
48.40
Save 3.75 (8%)
Catalog # Name Size Price (USD) Qty
A4192001 BenchStable™ DMEM/F12, 500 mL Each
66.65
Online Exclusive
71.75
Save 5.10 (7%)
A4192002 BenchStable™ DMEM/F12, 10 x 500 mL Each
550.65
Online Exclusive
596.00
Save 45.35 (8%)
10565018 DMEM/F-12, GlutaMAX™ supplement, 500 mL Each
65.65
Online Exclusive
71.75
Save 6.10 (9%)
NOTE: This supplement is suitable for mammalian cell cultures. It is not recommended for insect cell cultures.
Frequently asked questions (FAQs) for L-glutamine and GlutaMAX supplement
Would you like to learn more about using L-glutamine and GlutaMAX supplement in your cell culture studies?
Visit our L-glutamine and GlutaMAX FAQs
Product literature for L-glutamine and GlutaMAX supplement
Watch GlutaMAX Supplement video
Try the Gibco MediaSelect Tool
Easily find a media solution suited to your specific application. Use this interactive tool to select classical basal, protein expression, or viral vector media.
Your selections will allow you to choose from a catalog menu of media – or customize your own formulation to fit your experimental setup and budget. Where available, you can compare media formulations side-by-side.
Find your media with the Gibco MediaSelect Tool
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, helping to ensure the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Gibco Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.