research
Understanding disease progression and associated biomarkers is key to developing treatments for novel viruses. Even though patients may have varying immune responses, it’s possible to determine widespread prevention and treatment options. Thermo Fisher Scientific offers a complete portfolio of tools and technologies to enable SARS-CoV-2 virus research, supporting collaboration and the acceleration of lifesaving discoveries.
Pathogen biology
SARS-CoV-2, the virus responsible for COVID-19, is an enveloped, non-segmented, single-stranded positive-sense RNA (+ssRNA) virus and a newly identified β-coronavirus. The genome of ~30 kb consists of multiple open reading frames (ORFs) encoding four structural proteins: Spike (S), Envelope (E), Membrane (M), and Nucleocapsid (N), and a variety of non-structural proteins.
The SARS-CoV-2 viral lifecycle has yet to be resolved, a critical step to studying infection progression. Even though the replicative lifecycle of viruses differs greatly between species and categories, there are common stages that are essential for viral replication: attachment, entry and uncoating, replication, assembly, and release.
Untangling the viral replication pathways from host cell functions is challenging because viruses hijack cell machinery upon infection, creating cell conditions more conducive for virus production. Thermo Fisher Scientific offers an array of technologies and products to elucidate the basic stages of the viral life cycle and aid in the investigation of the biology of SARS-CoV-2. Some of these technologies and products are highlighted in the following sections.
Sample integrity is essential to the success of every storage approach.  So is the ability to easily share information designed to accelerate lifesaving discoveries.  Thermo Fisher Scientific offers the most diverse, cutting-edge portfolio of storage solutions and services designed to help you do both.  Our biobanking sample collection, preparation and automation equipment, cost storage solutions, wireless monitoring, tubes, vials, containers, reagents, and biobanking services give you the power to secure and preserve your samples, track your inventory, streamline productivity, and meet your toughest laboratory challenges.  At the same time, our informatics solutions enable you to share insights and information critical to advancing science and discovering the next big idea.
Lab equipment—reliable sample protection and preparation solutions with exceptional contamination protection, control and a wide range of options to fit your needs
BioBanking solutions—transforming life science research
Clinical research consumables—providing high quality and reliability to protect your samples and your staff
Gibco Viral Transport Medium Kit— to enable the safe transport and storage of samples for viral testing; suitable for the transport of specimens suspected to contain virus
RNase inhibitors—prevent degradation of RNA viruses by naturally-abundant RNases
Catalog # Name Size Price (USD) Qty
30APPRNIRR Sterilin Certified SPC Plain IRR Case of 400
310.80
Online Exclusive
354.00
Save 43.20 (12%)
SV200-101-LSB Smart-Vue™ 868MHz Wireless Radio Module System with external PT100 temperature probe,for use in ultra low freezers (Europe) Each -
7010 DNA AWAY Squeeze Bottle 250mL (8oz) Case of 12
501.59
Online Exclusive
567.60
Save 66.01 (12%)
7002 RNase AWAY™ Surface Decontaminant Case of 6
305.34
Online Exclusive
436.20
Save 130.86 (30%)
4672100BT E1-ClipTip Equalizer 8-ch 15-1250 μl BT Each
2,627.70
Online Exclusive
2,766.00
Save 138.30 (5%)
94420813 ClipTip 1250 Filter 8x96 Sterile pipette tips Case of 768
139.50
Online Exclusive
155.00
Save 15.50 (10%)
AM2694 SUPERase·In™ RNase Inhibitor (20 U/μL), 2500 U Each
218.00
10777019 RNaseOUT™ Recombinant Ribonuclease Inhibitor Each
255.00
AM7005 RNAsecure™ RNase Inactivation Reagent, 1 mL Each
57.65
Online Exclusive
61.25
Save 3.60 (6%)
N8080119 RNase Inhibitor Each
131.00
To learn more about isolation of viral RNA for SARS-CoV-2 research, visit the Sample Prep section on the Epidemiological Research page.
Nucleic acid quantification and qualification for SARS-CoV-2
By absorbance
  • Quantify DNA, RNA, and protein in seconds using only 1–2 µL of sample—no need for dilution. Thermo Scientific NanoDrop One instruments are built with Thermo Scientific Acclaro Sample Intelligence technology to help you determine if your sample is suitable for downstream applications.
  • Prevent costly delays—Acclaro software identifies contaminants (protein, phenol, guanidine salts) and provides corrected concentrations to ensure experimental success
  • Quickly collect data—save time with Auto-Blank and Auto-Measure modes
  • Troubleshoot results in real-time—information alerts trigger guided technical support for fast answers
By fluorescence
The Invitrogen Qubit Fluorometer is designed to quickly and specifically quantify DNA or RNA.
  • Qubit assay dyes bind selectively to DNA, RNA, or protein, making it orders of magnitude more sensitive than UV absorbance
  • Uses as little as 1 μL of sample, even with very dilute samples
  • Fast, reliable detection of degraded RNA with the Invitrogen Qubit RNA IQ Assay
Compare nucleic acid quantitation and qualification technologies ›
Understand variation in the SARS-CoV 2 genome and how that information can be used to track community spread.
 AmpliTaq Gold DNA PolymerasePlatinum II Taq Hot-Start DNA Polymerase
Compatible with TaqMan probesYesYes
Cycling time60 sec/kb 15 sec/kb 
PCR sensitivity +++++
Universal annealing temperature  NoYes
Learn more about cloning of viral genes ›
Catalog # Name Size Price (USD) Qty
4398823 AmpliTaq Gold™ 360 DNA Polymerase, 250 Units Each
409.65
Online Exclusive
431.00
Save 21.35 (5%)
4398881 AmpliTaq Gold™ 360 Master Mix, 5 mL Each
471.00
14966005 Platinum™ II Taq Hot-Start DNA Polymerase, 500 reactions Each
661.65
Online Exclusive
688.00
Save 26.35 (4%)
14000013 Platinum™ II Hot-Start PCR Master Mix (2X), 200 reactions Each
389.65
Online Exclusive
412.00
Save 22.35 (5%)
14001013 Platinum™ II Hot-Start Green PCR Master Mix (2X), 200 reactions Each
396.65
Online Exclusive
413.00
Save 16.35 (4%)
For viruses to access intracellular sites of replication in the cytosol or nucleus, cellular membranes present the first barrier for entry into the host cell. Viral-encoded envelope or capsid proteins serve as binding partners for host cell molecules or receptors, and their binding specificity defines the cell and tissue tropism, infectivity, and thus the nature of disease.
Virus neutralization
Traditional plaque assays are being replaced  by neutralization assays, such as the plaque reduction neutralization test (PRNT) to test antibodies that neutralize and prevent virions from infecting cultured cells. PRNT is now required by the Centers for Disease Control (CDC) for some viruses utilizing high-content screening instrumentation to measure viral infectivity and neutralization (1). Below are high-content screening instrumentation options for PRNT assays and other high throughput applications.
ProductCat. No.
CellInsight CX5 PlatformRequest a quote ›
CellInsight CX7 PlatformRequest a quote ›
CellInsight CX7 LZR PlatformRequest a quote ›
Learn more about Invitrogen high-content screening platforms ›
SARS-CoV-2 receptors and other entry factors
The entry of coronaviruses into target host cells has been shown to be facilitated by the Spike (S) protein, with the SARS-CoV-2 S1 subunit of the spike protein critical for binding to the cell surface receptor angiotensin-converting enzyme 2 (ACE2). Priming of the S protein by the cellular serine protease, TMPRSS2, generates the S2 subunit required for fusion of the viral and cellular membrane, facilitating entry of the virus into the host cell (2).
Fully characterizing the molecules and signaling pathways implicated in SARS-CoV-2 entry is key to understanding infectivity, tropism, pathogenesis, and immune evasion.
  1. Human Coronavirus Entry Factor Panel
  2. Mouse Coronavirus Entry Factor Panel
  3. Rat Coronavirus Entry Factor Panel
Tracking internalization
The entry mechanism of coronaviruses has been shown to be mediated via the endocytic pathway. The exact mechanism of endocytosis has yet to be determined, with reports indicating both clathrin-dependent and –independent endocytosis (3). Typically, internalized viruses are directed through the endosomes to the lysosome. Viral uncoating is a highly variable process that occurs at different stages after viral entry and may be influenced by pH, lipid composition, proteolytic cleavage, and redox reactions among other factors (4).
Thermo Fisher Scientific offers an extensive portfolio of Invitrogen cell imaging products and technologies to help researchers track the internalization of SARS-CoV-2 viral particle.
Catalog # Name Size Price (USD) Qty
P35373 pHrodo™ Green AM Intracellular pH Indicator, 50 μL Each
595.65
Online Exclusive
614.00
Save 18.35 (3%)
P35372 pHrodo™ Red AM Intracellular pH Indicator, 50 μL Each
598.65
Online Exclusive
620.00
Save 21.35 (3%)
P35380 pHrodo™ AM Variety Pack Each
284.00
P35379 Intracellular pH Calibration Buffer Kit Each
313.65
Online Exclusive
327.00
Save 13.35 (4%)
Imaging cell structure
Viral spike proteins reside on the surface of viruses and are used to bind to host cells, enabling viral infection. Knowledge of the precise, 3D structure of these proteins is essential for the generation of specific, effective drugs and vaccines. It also provides general insight into how the virus evades the immune system.
Learn more Invitrogen probes for imaging cell structure ›
Virus-host cell interaction
In order for a virus to be effective, it needs to hijack the host cell and replicate. Therefore, the interaction of the virus with host cell proteins is a crucial part of this strategy. This interaction not only ensures the replication of the virus, but it is also a strategy for the virus to evade the defense mechanisms initiated by the host cells.
Examining the virus-host cell protein interactions can provide insights into processes such as viral entry, virus genome replication within the host cell, and the ways the viral genome can spread across the cell. Additionally, examining virus-host cell protein interactions could yield valuable information and help scientists develop broadly acting antiviral therapeutics. 
Learn more about Viral proteomics and metabolomics mass spectrometry ›
References
  1. Gates I, Olson V, Smith S, Patel N, Damon I, Karem K. Development of a High-Content Orthopoxvirus Infectivity and Neutralization Assays. PLoS One. 2015;10(10):e0138836.
  2. Hoffmann M, Kleine-Weber H, Schroeder S, et al. SARS-CoV-2 Cell Entry Depends on ACE2 and TMPRSS2 and Is Blocked by a Clinically Proven Protease Inhibitor. Cell. 2020;181(2):271‐280.e8.
  3. Yang N, Shen HM. Targeting the Endocytic Pathway and Autophagy Process as a Novel Therapeutic Strategy in COVID-19. Int J Biol Sci. 2020;16(10):1724‐1731.
  4. Yamauchi Y, Helenius A. Virus entry at a glance. J Cell Sci. 2013;126(Pt 6):1289‐1295.
Recombinant Proteins for SARS-CoV-2 research
Real-time PCR solutions for SARS-CoV-2 research
Explore sample prep reagents, assays, master mixes, instrumentation, and analysis software for real-time PCR applications in SARS-CoV-2 research.
Catalog #Assay IDNameSize
4331182Vi06439674_s1CoV_HKU1250 rxns
4331182Vi06439646_s1CoV_OC43250 rxns
4331182Vi06439673_s1CoV_NL63250 rxns
4331182Vi06439671_s1CoV_229E250 rxns
4331182Vi06439644_s1MERS_CoV250 rxns
4331182Vi06439634_s1SARS_CoV250 rxns
4331182Hs01085333_m1 Human ACE2250 rxns
4331182Hs01122322_m1 Human TMPRSS2250 rxns
4331182Hs00947439_m1Human CTSB250 rxns
4331182Hs00964650_m1 Human CTSL250 rxns
4331182Hs00174265_m1 Human ANPEP250 rxns
4331182Hs00897386_m1Human DPP4250 rxns
4331182Hs00949382_m1 Human ST6GAL1250 rxns
4331182Hs00920870_m1 Human ST3GAL4250 rxns
To order, search the Assay ID or Name using our ordering tool.    Ordering tool
Catalog # Name Size Price (USD) Qty
A47695 CTS™ Rotea™ Counterflow Centrifugation System + 2-Year Service + IQOQ Each -
A1067501 StemPro™ MSC SFM XenoFree Each
521.90
Special offer
Online exclusive
Ends: 21-Jun-2025
614.00
Save 92.10 (15%)
A1033201 StemPro™ MSC SFM, 1 Kit Each
472.82
Special offer
Online exclusive
Ends: 21-Jun-2025
528.00
Save 55.18 (10%)
A1371301 CTS™ Synth-a-Freeze™ Medium Each
249.00
A1286001 CTS™ GlutaMAX™-I Supplement, 100 mL bottle Each
71.89
Special offer
Online exclusive
Ends: 21-Jun-2025
79.00
Save 7.11 (9%)
A4132602 CTS™ LV-MAX™ Transfection Kit, 1 Kit (For 10 L of LV Production) Each
19,920.00
CMAX00001 Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent, 0.1 mL Each
116.65
Online Exclusive
121.00
Save 4.35 (4%)
A36496 TrueCut™ Cas9 Protein v2, 10 μg Each
90.65
Online Exclusive
92.75
Save 2.10 (2%)
4460623 MycoSEQ™ Mycoplasma Detection Kit, 100 Reactions Each
4,195.00
The uncoating and release of viral RNA into the cytosol results in the translation of viral proteins from the six or more open reading frames (ORFs) identified in coronaviruses. Translation begins at ORF1 a/b, which encodes a large polyprotein that is cleaved to generate at least 16 nonstructural proteins (nsps) that are involved in viral replication (1,2). These nsps include replicases that are involved in generating additional full-length genomes (1). The remaining ORFs encode for four structural proteins: Spike (S), Envelope (E), Membrane (M), and Nucleocapsid (N), and several accessory proteins (1).
Branched DNA Signal Amplification
One approach to detection of SARS-CoV-2 RNA is performed through branched DNA (bDNA) signal amplification technology. The presence of viral RNA transcripts in infection models can be confirmed through bDNA technology in imaging or flow cytometry using the Invitrogen ViewRNA Assay or Invitrogen PrimeFlow RNA Assay, respectively.
Real-time PCR and PCR
Use real-time PCR to detect and quantify SARS-CoV-2 RNA in different tissues, cultures, or specimens to elucidate fundamentals of pathogen biology including replication kinetics. We offer predesigned research assays for the SARS-CoV-2S gene, or you can design custom primers and probes for other viral targets.
Learn more about real-time PCR solutions for SARS-CoV-2 research.
For RNA viral studies that require reverse transcription (RT), such as 2-step real-time PCR or clone development, researchers can rely on SuperScript IV, our newest RT enzyme. Reliability and performance of SuperScript IV Reverse Transcriptase products help facilitate:
For RNA viral studies that require reverse transcription (RT), researchers can rely on SuperScript IV, our newest RT enzyme. Reliability and performance of SuperScript IV Reverse Transcriptase products help facilitate:
  • Superior sensitivity to detect RNA input amounts down to 0.01 pg total RNA
  • 10 minute RT reaction time for fast performance
  • Significantly better resistance to contaminating inhibitors
See how the enzyme was used in a SARS-CoV-2 research study:
Xie X, Muruato A, Lokugamage KG, et al. (2020) An Infectious cDNA Clone of SARS-CoV-2.Cell Host Microbe S1931-3128(20)30231-6.
References:
1. Perlman S, Netland J. Coronaviruses post-SARS: update on replication and pathogenesis. Nat Rev Microbiol. 2009;7(6):439‐450.
2. Kumar S., Nyodu R., Maurya V.K., Saxena S.K. (2020) Morphology, Genome Organization, Replication, and Pathogenesis of Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2). In: Saxena S. (eds) Coronavirus Disease 2019 (COVID-19). Medical Virology: From Pathogenesis to Disease Control. Springer, Singapore
Catalog # Name Size Price (USD) Qty
18090200 SuperScript™ IV Reverse Transcriptase, 4 x 10,000 Units Each
1,763.65
Online Exclusive
1,876.00
Save 112.35 (6%)
N8080127 Random Hexamers (50 μM) Each
85.40
Special offer
Online exclusive
Ends: 01-Jun-2025
122.00
Save 36.60 (30%)
18418020 Oligo(dT)20 Primer Each
85.40
Special offer
Online exclusive
Ends: 01-Jun-2025
122.00
Save 36.60 (30%)
18427089 dNTP Mix (10 mM ea), 5 x 1 mL Each
1,290.65
Online Exclusive
1,418.00
Save 127.35 (9%)
10777019 RNaseOUT™ Recombinant Ribonuclease Inhibitor Each
255.00
10813012 UltraPure™ DEPC-treated Water, 4 x 1.25 mL Each
82.65
Online Exclusive
90.00
Save 7.35 (8%)
14000013 Platinum™ II Hot-Start PCR Master Mix (2X), 200 reactions Each
389.65
Online Exclusive
412.00
Save 22.35 (5%)
4452300 Veriti™ Dx 96-well Thermal Cycler, 0.2 mL Each -
AMF7000 EVOS™ M7000 Imaging System Each -
18090050 SuperScript™ IV Reverse Transcriptase, 10,000 Units Each
497.65
Online Exclusive
530.00
Save 32.35 (6%)
12594025 SuperScript™ IV One-Step RT-PCR System, Each Each
396.65
Online Exclusive
429.00
Save 32.35 (8%)
Real-time PCR solutions for SARS-CoV-2 research
Explore sample prep reagents, assays, master mixes, instrumentation, and analysis software for real-time PCR applications in SARS-CoV-2.
TaqMan Gene Expression Assays
Catalog #Assay IDNameSize
4331182Vi06439674_s1CoV_HKU1250 rxns
4331182Vi06439673_s1CoV_NL63250 rxns
4331182Vi06439671_s1CoV_229E250 rxns
4331182Vi06439644_s1MERS_CoV250 rxns
4331182Vi06439634_s1SARS_CoV250 rxns
To order, search the Assay ID or Name using our ordering tool.    Ordering tool
The assembly of the coronavirus particle starts with the helical nucleocapsids, which are formed by the interaction of newly synthesized viral genomic RNA with the nucleocapsid phosphoproteins (1). The triple-spanning membrane glycoprotein M is translated on membrane-bound polysomes and co-translationally inserted into the membrane of the endoplasmic reticulum (ER) and transported to the ER-Golgi intermediary compartment (ERGIC). At the ERGIC, the helical nucleocapsid interacts with the C-terminal domain of the M protein, which is exposed to the cytosol (2). In a process thought to be mainly driven by its transmembrane domain, M oligomerization leads to the formation of lattice structures into which the S and E membrane proteins are integrated (1,2). The structure is then transported by secretory vesicles to the plasma and released. The final coronavirus particle consists of the helical nucleocapsid surrounded by a lipid layer consisting of inserted spike (S), membrane (M), and envelope (E) proteins (2).
Expi293 Expression System for structural biology applications—a suite of products that enables the study of viral surface proteins by providing high titers of uniformly glycosylated proteins, the ability to modulate expression levels of proteins, and protein labeling kits for streamlined studies by X-ray crystallography or Cryo-EM
References:
1. Tseng YT, Wang SM, Huang KJ, Lee AI, Chiang CC, Wang CT. Self-assembly of severe acute respiratory syndrome coronavirus membrane protein. J Biol Chem. 2010;285(17):12862‐12872. 
2. Siu YL, Teoh KT, Lo J, et al. The M, E, and N structural proteins of the severe acute respiratory syndrome coronavirus are required for efficient assembly, trafficking, and release of virus-like particles.J Virol. 2008;82(22):11318‐11330. 
The structure of the virus has a profound effect on its interaction with host cells and how it functions during its lifecycle. In general, viruses are composed of RNA or DNA surrounded by a capsid composed of proteins and glycoproteins. The capsid can vary depending upon the virus, but its primary goal is to protect the viral genome. Some viruses are further encompassed by membrane envelopes composed of lipids and proteins. The surface of the envelope has additional proteins, specifically glycoproteins, which are involved in the process of binding to the host cells for viral entry. A number of mass spectrometry tools are available for the characterization of the different parts of the SARS-CoV-2 virus.
Learn more about Glycoproteomics, Glycomics, Native MS, HDX-MS ›
Sanger Sequencing Solutions for SARS-CoV-2 Research
Sanger_sequencing_SARS-CoV-2_research
Generate highly specific targeted results
Sanger sequencing can be completed in less than one workday—from sample to answer—helping to make your research of SARS-CoV-2 (the coronavirus that causes COVID-19) fast and cost-effective.  
Download detailed protocols for single-sample sequencing of the virus to identify mutations and to confirm data from NGS sequencing.
Learn More ›
Host response
The host response to SARS-CoV-2 infection, from host pathogen interactions and signaling events to innate and adaptive mediated immune responses, plays a critical role in studies of human health implications. Currently, treatment of SARS-CoV-2 infection is palliative, with the primary cause of mortality being respiratory failure from acute respiratory distress syndrome (ARDS) (1).  Recent studies show that management of the immune-mediated inflammatory cytokine response may be beneficial. In severe infection this response is often referred to as cytokine release storm or syndrome (CRS), which can lead to hyper-inflammation, multi-organ failure, and eventually death (1). Due to the wide range of symptoms there is research evidence that the immune response factors vary greatly among subjects, providing some with stronger early defenses that never progress to severe cases. It is therefore critical to understand the cellular and biochemical differences in the innate and adaptive immune response in patients that are asymptomatic, have mild symptoms, or have severe CRS (2,3).
From large scale genomics and proteomics to more targeted approaches, Thermo Fisher Scientific offers researchers an extensive portfolio of solutions for immunological and infectious disease studies.
Maintaining the integrity of your samples at every point from collection to storage is imperative to obtaining quality sample and associated data. This begins with the preparation of biological samples, which lays the groundwork for a range of critical research objectives such as drug discovery, disease research and beyond. Our solutions support sample preparation, from extraction to fractionation and aliquoting, while maintaining traceability from the point of collection at a range of throughput levels for biobanks at any scale. The safety and security of your samples is non-negotiable. We understand the dual importance of sample integrity and inventory management and offer a comprehensive sample storage solution designed to protect your samples, track your inventory, and provide peace of mind.
The identification of molecular markers or molecular signatures of biological processes using high throughput technologies capable of comprehensive analysis of genomic and other biological molecules has provided significant insights and helped with understanding of host response to infection and disease progression.
Hypothesis-free target identification and evaluation of host gene expression is achieved with higher accuracy, in less time, using Invitrogen Collibri Library Prep Kits for Illumina systems.
  • Increase the quality of sequences to obtain the entire genome
  • Improve accuracy and speed of RNA sequencing results by adding adapters for Illumina NGS directly to RNA
  • Identify differences in gene expression that may offer insight into why some people become gravely ill and others do not show symptoms
Complete SARS-CoV-2 genome sequencing with Ion Torrent NGS
Ion Torrent targeted next-generation sequencing (NGS) enables a streamlined research workflow for complete genome sequencing and epidemiological studies of SARS-CoV-2. Through the use of a set of highly specific, universal coronavirus primers in combination with a high-fidelity master mix, all genomic segments are amplified and the DNA amplicons are sequenced on any Ion Torrent system to deliver highly accurate coronavirus typing in under a day. The Ion AmpliSeq SARS-CoV-2 Research Panel is compatible with the Ion GeneStudio S5 and Ion Torrent Genexus systems.  In addition to the Ion AmpliSeq SARS-CoV-2 Research Panel workflow, which covers >99% of the viral genome and variants, Ion Torrent GeneStudio systems can be used for de novo (whole genome) sequencing of SARS-CoV-2 .
Sanger sequencing to confirm partial and full genome SARS-CoV-2 sequence following NGS
Sanger sequencing is a simple, fast, and cost-effective orthogonal method to confirm SARS-CoV-2 identified by NGS or by RT-PCR.  It was used in the connection of viral contigs to generate the first full length genome sequence of the novel coronavirus SARS-CoV-2.
The entire workflow can be completed, from sample to answer, in less than one workday.  Thermo Fisher Scientific offers products to support the entire workflow from PCR amplification to data analysis.  The SeqStudio Genetic Analyzer is an affordable, low-throughput, cartridge-based benchtop genetic analyzer that eliminates complex instrument setup to deliver Sanger sequencing runs in as little as 30 minutes. 
Learn more about Sanger sequencing solutions for SARS-CoV-2 research ›
Understand how genomic variation can impact response to pathogens
Learn more about T-cell and B-cell receptor variation, as well as how gut microbiome can influence immune response to SARS-CoV-2.
Understand variation in the SARS-CoV 2 genome and how that information can be used to track community spread.
Learn more about the Ion AmpliSeq SARS-CoV-2 Research Panel or request project costs ›
While clinical guidance issued by the CDC currently lists laboratory markers that are associated with increased COVID-19 illness severity, including but not limited to elevated liver enzymes, lymphopenia, and neutropenia (1), there is an immediate need by researchers to identify and develop other biomarkers to understand the biology of the host response to the infection. These include biomarkers to monitor cellular response, including the innate sensing of viral infections, host susceptibility factors, the activation of anti-viral pathways, and induction of pro-inflammatory responses, to help characterize the molecular networks underlying symptomatic infections and the progression and pathogenesis of the infections.
Real-time PCR for pathway and biomarker characterization
Real-time PCR for SARS-CoV-2 research has multiple applications in biomarker and disease-pathway research, including gene expression and miRNA analysis. Studying differentially expressed genes in phenotypically different SARS-CoV-2 samples can help researchers identify or confirm novel biomarkers and potential therapeutic targets. Current research also indicates that miRNA expression may play a key role in disease pathogenesis. TaqMan Advanced miRNA Assays provide the highest level of sensitivity and specificity for miRNA profiling and screening studies, detecting down to 60 miRNA copies into the cDNA.
Learn more about gene expression analysis solutions for SARS-CoV-2 research ›
Learn more about miRNA and noncoding RNA analysis solutions for SARS-CoV-2 research ›
2D and 3D cell culture model for biomarker research
Researchers rely upon 2D cell culture models that are easy to use and well-established for studying infectious diseases. Thermo Fisher Scientific provides cell culture products for basic research to specialty assays. These include cell culture media, high activity growth factors, fetal bovine serum, and tissue culture grade plastics for imaging or various cell attachment needs.
Cardiovascular, pulmonary, hematological, and neurological complications in the context of SARS-CoV-2 infection emphasize the importance of establishing relevant physiological models to understand underlying cellular mechanisms. 3D cellular models like organoids and spheroids offer an opportunity to better understand complex biology and have shown great potential in recent publications (1,2). Explore end-to-end solutions with Thermo Fisher 3D cell culturing products.
Contact cell model services about your project today ›
Discover tools, protocols and product solutions to produce your 3D model, including the Nunclon Sphera 3D culture system ›
3D cell culture
Catalog # Name Size Price (USD) Qty
A4192101 BenchStable™ DMEM, 500 mL Each
44.65
Online Exclusive
48.37
Save 3.72 (8%)
A4192001 BenchStable™ DMEM/F12, 500 mL Each
66.65
Online Exclusive
71.75
Save 5.10 (7%)
A4192301 BenchStable™ RPMI 1640, 500 mL Each
45.65
Online Exclusive
49.31
Save 3.66 (7%)
10091148 Fetal Bovine Serum, qualified, New Zealand, 500 mL Each
1,078.00
10099141 Fetal Bovine Serum, qualified, Australia, 500 mL Each
1,062.00
16000044 Fetal Bovine Serum, certified, United States, 500 mL Each -
174932 6 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
169.65
Online Exclusive
181.00
Save 11.35 (6%)
174930 24 Well Plate, Sphera Low-Attachment Surface, Pack of 1 Case of 7
164.65
Online Exclusive
177.00
Save 12.35 (7%)
174929 96 Well Round (U) Bottom Plate, Sphera Low-Attachment Surface, Pack of 5 Case of 50
1,090.65
Online Exclusive
1,192.00
Save 101.35 (9%)
Cellular analysis using imaging and western blotting
In order to study pathways and biomarkers in 2D or 3D cellular models in the context of SARS-CoV-2 infection, commonly used techniques such as imaging and western blotting can provide spatiotemporal and quantitative analysis of protein expression. For example, Lamers et al. studied 3D airway and gut organoids in the context of SARS-CoV-2 infection to understand the general viral response utilizing Thermo Fisher imaging reagents for mounting specimens, performing nuclear and cytoskeletal staining, and antibody based detection for specific biomarkers (1).
To detect biomarkers in cell or tissue lysates, Thermo Fisher Scientific provides end to end western blotting solutions, or use this convenient blot builder tool to help determine the correct western workflow products to fit your experimental needs.
Gene expression analysis to identify biomarkers for infection severity
Recent publications have shown host response to pathogen infection to be as important as viral strain sequence differences in understanding human health implications of SARS-CoV-2 infection The Ion AmpliSeq Transcriptome Human Gene Expression Kit and Ion AmpliSeq Transcriptome Mouse Gene Expression Kit are comprehensive tools for gene-level expression analysis. Targeting >20,000 human or mouse RefSeq transcripts in a single reaction with an FFPE-compatible workflow, the complete Ion AmpliSeq Transcriptome solution is compatible with existing downstream microarray or next-generation sequencing (NGS) analysis pipelines for fast and straightforward data interpretation.
QuantiGene Plex assays provide a fast and high-throughput solution for multiplexed gene expression quantitation, allowing the simultaneous measurement of up to 80 genes of interest in a single well of a 96- or 384-well plate. The QuantiGene Plex assay is hybridization-based and incorporates branched DNA (bDNA) technology, which uses signal amplification rather than target amplification for direct measurement of RNA or DNA transcripts. The assay is extremely easy to use with a simple ELISA-like workflow, and does not require RNA purification. It works with difficult sample types including degraded and cross-linked RNA in FFPE tissues and directly with blood, with no RNA purification required. QuantiGene Singleplex assays rely on luminescence readouts and can be analyzed on the Varioskan Plate Reader models, while the QuantiGene Plex assay uses the Luminex platform.
Immune response and microbiome biomarker identification
Understanding of the immune repertoire through T-cell and B-cell receptor sequencing in addition to gut microbiome profiles can identify important biomarkers for understanding response to SARS-CoV-2.
References:
1. Lamers MM, Beumer J, van der Vaart J, et al. SARS-CoV-2 productively infects human gut enterocytes [published online ahead of print, 2020 May 1]. Science. 2020;eabc1669.
2. Yang L, Han Y, Nilsson-Payant BE, et al. A Human Pluripotent Stem Cell-based Platform to Study SARS-CoV-2 Tropism and Model Virus Infection in Human Cells and Organoids. Cell Stem Cell. 2020;27(1):125-136.e7.
Multi-dimensional host response research with Ion Torrent Next-Generation Sequencing (NGS)
Recent publications have shown host response to pathogen infection to be as important as viral strain sequence differences in understanding human health implications of SARS-CoV-2 infection. Deeper understanding of the adaptive immune system, inflammation markers, and the human microbiome powers better insights into its role in health and response to pathogens, leading to crucial developments in future treatments such as immunotherapies. The Ion Torrent SARS-CoV-2 research portfolio enables a multi-dimensional research approach to repertoire sequencing and immune response, in addition to end to end workflows for broader transcriptome sequencing or human microbiome analysis. Each targeted next-generation sequencing (NGS) assay is powered by the accuracy and sensitivity of Ion Torrent technology to help discover more from precious samples along with highly automated end to end workflows including robust informatics tools. Combining host response with epidemiological studies, the Ion Torrent SARS-Cov-2 research portfolio presents a rapid and scalable way to address multiple facets of infectious disease research.
For human research studies:
For mouse studies:
The Ion GeneStudio S5 System is a scalable, targeted-NGS workhorse with wide research application breadth and throughput capability and is compatible with our complete portfolio of immune response assays.
Learn more about the Ion GeneStudio S5 System or request a quote ›
QuantiGene gene expression assays for biomarker verification
The Invitrogen QuantiGene Plex assays provide an accurate and precise method for single or multiplexed targeted biomarker gene expression quantitation. The QuantiGene assay is also available as a single plex assay, for the measurement of one gene target per well, while the QuantiGene Plex assay is used for multiplexed gene expression quantitation and can be used to measure up to 80 gene targets per well. QuantiGene Singleplex assays utilize luminescence detection enabled by Varioskan Plate Readers, while the QuantiGene Plex assay relies on the Luminex platform.
ProductCat No.
QuantiGene SingleplexRequest a quote ›
QuantiGene PlexRequest a quote ›
Targeted sequencing
Several methods have been developed for the targeted enrichment of genomic DNA for a variety of clinical and research applications. Typically, they are based on a multiplexed PCR amplification reaction and DNA hybridization. All these DNA enrichment methods rely heavily on fragmentation of genomic DNA prior to amplification, resulting in relatively short (less than 1000 base-pair) sequencing templates.
Targeted sequencing workflow implementing target captured by hybridization includes several general steps - sample preparation, library construction, target enrichment, sequencing and analysis.
Select products for targeted sequencing workflow
Catalog # Name Size Price (USD) Qty
AM1980 GLOBINclear™-Human Kit, for globin mRNA depletion Each
728.00
AM1981 GLOBINclear™ Kit, mouse/rat, for globin mRNA depletion Each
710.00
15401011 Yeast tRNA, 25 mg Each
224.65
Online Exclusive
249.00
Save 24.35 (10%)
15401029 Yeast tRNA, 50 mg Each
278.65
Online Exclusive
307.00
Save 28.35 (9%)
AM8740 RNA Fragmentation Reagents Each
123.00
15279011 Human Cot-1 DNA™ Each
355.65
Online Exclusive
391.00
Save 35.35 (9%)
18440016 Mouse Cot-1 DNA Each
377.65
Online Exclusive
407.00
Save 29.35 (7%)
15279101 Human Cot-1 DNA™-Fluorometric QC Each
609.65
Online Exclusive
662.00
Save 52.35 (8%)
19524016 Biotin-14-dATP Each
644.65
Online Exclusive
712.00
Save 67.35 (9%)
19518018 Biotin-14-dCTP Each
586.65
Online Exclusive
640.00
Save 53.35 (8%)
19519016 Biotin-14-CTP Each
686.00
18094011 BioPrime™ DNA Labeling System Each
702.65
Online Exclusive
768.00
Save 65.35 (9%)
18095012 BioPrime™ Array CGH Genomic Labeling Module Each
728.00
18010025 DNA Polymerase I, 1000 U Each
533.65
Online Exclusive
638.00
Save 104.35 (16%)
18010017 DNA Polymerase I, 250 U Each
216.65
Online Exclusive
237.00
Save 20.35 (9%)
The host’s immune system is its defense system against viral infections. The characterization of the various molecules involved in the immune systems response to viruses is necessary for the development of treatments that can be used to suppress viral replication and propagation, as well as alleviate any symptoms of diseases caused by the virus. Mass spectrometry characterization and quantitation of proteins and metabolites can provide important information about how the immune system functions. Learn more ›
Flow cytometry analysis of intracellular and cell-surface targets can help elucidate a more complete mechanism of the immune response to viral infections. Cossarizza et al. demonstrated guidelines to safely analyze samples for intracellular cytokine detection and relative differences among immune cells (1). Important cytokine targets, such as those listed in the table below, as well as cytotoxic markers Granzyme A, B, K and M, Perforin, and Granulysin, can be analyzed using Invitrogen flow cytometry antibodies with the Invitrogen Attune NxT flow cytometer to understand the immune response during SARS-CoV-2 infection (1,2). The targets listed below can also be analyzed using immunoassays found in the following section.
TargetDistributionFunction
IFNgammaCytotoxic T cells, NK cells
Viral and parasitic responses
TNFalpha
Macrophages
Viral and parasitic responses, regulation of immune cells
IL-6
Activated T cells
Viral responses
IL-10
Macrophages, Th2 cells
Enhances antibody production, anti-inflammatory cytokine
IL-17
Th17 cells
Pro-inflammatory cytokine
Granzyme A
Cytotoxic T cells, NK cells
Target cell apoptotic lysis, cell-mediated immune responses
Granzyme B
Cytotoxic T cells, NK cells
Target cell apoptotic lysis, cell-mediated immune responses
Granzyme K
Cytotoxic T cells, NK cells
Target cell apoptotic lysis, cell-mediated immune responses
Granzyme M
Cytotoxic T cells, NK cells
Target cell apoptotic lysis, cell-mediated immune responses
Granulyisn
Cytotoxic T cells, NK cells
Target cell apoptotic lysis, cell-mediated immune responses
Perforin
Cytotoxic T cells
Viral response, pore forming
PD-1
Activated T cells
Immune inhibitory molecule
Tim-3
Th1, Th17, Tregs, CD8+ T cell subsets
Immune inhibitory molecule
In addition, Invitrogen offers out-of-the-box solutions such as the essential phenotyping kits, which are efficient and cost-effective methods for flow cytometry-based analysis. These ready-to-use kits have been designed for the reliable identification of T-cell subsets including CD4+, CD8+, regulatory T-cells (Tregs), and T helper cell subsets in human blood on any flow cytometer equipped with violet 405 nm, blue 488 nm, and red 633 nm lasers, such as the Invitrogen Attune NxT flow cytometer. If you need a larger custom panel to discover more about your leukocyte lineages, request a quote below or build your own panel using the Invitrogen Flow Cytometry Panel Builder.
Find antibodies for flow cytometry ›
References:
1. Cossarizza A, Gibellini L, De Biasi S, et al. Handling and Processing of Blood Specimens from Patients with COVID-19 for Safe Studies on Cell Phenotype and Cytokine Storm [published online ahead of print, 2020 Apr 10]. Cytometry A. 2020;10.1002/cyto.a.24009.
2. Thevarajan I, Nguyen THO, Koutsakos M, et al. Breadth of concomitant immune responses prior to patient recovery: a case report of non-severe COVID-19. Nat Med. 2020;26(4):453‐455.
Immunological assays can help pinpoint cellular subsets and associated secreted molecules important for viral infection and host immunology. They are used to understand virulence and attenuation and to develop neutralizing vaccines.
Invitrogen immunoassay kits are easy to use for quantification of inflammatory-related cytokines and chemokines in serum, plasma, or other biological fluids.
ELISA for single analyte detection
Measure key inflammatory cytokines and chemokines such as IFNγ, IL-1B, IL-2, IL-6, GM-CSF, IP-10, MCP-1, MIP-1a, and TNFα or acute phase indicators such as Ferritin or CRP.
See the complete list of ELISA menu selection.
Catalog # Name Size Price (USD) Qty
BMS213INST Human IL-6 Instant ELISA™ Kit 128 tests
695.65
Online Exclusive
736.00
Save 40.35 (5%)
BMS288INST Human CRP Instant ELISA™ Kit 128 tests
689.65
Online Exclusive
732.00
Save 42.35 (6%)
EHFTL Human Ferritin ELISA Kit 96 tests
638.65
Online Exclusive
660.00
Save 21.35 (3%)
VL0LATD0 Varioskan™ LUX multimode microplate reader, Absorbance, Fluorescence intensity, Luminescence, Time-resolved fluorescence and AlphaScreen Each
35,040.00
Immunoassay plates are available with a variety of surfaces optimized to capture the biomolecule of your choice. To help identify the best surface for your application see our  immunoassay plate guide.
Serological assays
Serological blood assays can detect antibodies that remain in the blood after SARS-CoV-2 infection. This can enable determination of convalescent plasma samples, or those that contain SARS-CoV-2 specific antibodies. Search for anti-human IgG or IgM, or plate reader-based detection below:
Find anti-human antibodies for use in serological testing (lateral flow or ELISA) ›
See plate reader instrumentation for fluorescence and luminescence readouts with ELISA, serological assays ›
Advanced qPCR-based immunoassay for fast, high-sensitivity detection
ProQuantum high-sensitivity immunoassays enable detection of inflammatory markers in only two hours. No wash steps minimize contaminated waste. Run on any qPCR instrument, the assay provides a large dynamic range to easily measure low and elevated protein levels while only consuming 2 µL of serum sample.
Learn more and see the complete list of ProQuantum targets available ›
Catalog # Name Size Price (USD) Qty
A35573 Human IL-6 ProQuantum Immunoassay Kit 96 tests
718.00
A35574 Human IL-1 beta ProQuantum Immunoassay Kit 96 tests
643.65
Online Exclusive
710.00
Save 66.35 (9%)
A35601 Human TNF alpha ProQuantum Immunoassay Kit 96 tests
664.65
Online Exclusive
718.00
Save 53.35 (7%)
ProcartaPlex multiplexed immunoassays
ProcartaPlex multiplex immunoassays use the Luminex xMAP (multianalyte profiling) technology that enables the simultaneous detection and quantitation of up to 65 protein targets in a single 25–50 µL sample of plasma, serum, cell culture supernatants, or other bodily fluids. In the case of SARS-CoV-2 infection, ProcartaPlex immunoassays provide customizable panels to study Cytokine Release Storm (21 preselected targets commonly found in literature), or use our pre-configured panels for your viral research.
Recombinant proteins
Our recombinant proteins are low-endotoxin and validated via bioassay and SDS-PAGE. They are provided in a carrier-free format ideal for cell culture or production of standards for ELISA or western blotting.
Find your protein of interest ›
Analyze host immune response with real-time PCR
Interrogating the differences in the innate and adaptive immune responses in patients that are asymptomatic, mildly symptomatic, or acutely ill is critical to understand disease pathways and possible therapeutic targets. Our real-time PCR solutions for SARS-CoV-2 research enable scientists to investigate differential gene expression of cytokines, chemokines, and growth factors, identify host genetic determinants that impact the immune response, or monitor miRNA expression to better understand the role of these molecules in antiviral responses. Monitoring expression of antiviral host restriction factors is also a key component of understanding how host cells restrict entry, inhibit viral RNA synthesis, and block viral translation.
  1. Mehta P, McAuley DF, Brown M, et al. COVID-19: consider cytokine storm syndromes and immunosuppression. Lancet. 2020;395(10229):1033‐1034.
  2. Huang C, Wang Y, Li X, et al. Clinical features of patients infected with 2019 novel coronavirus in Wuhan, China [published correction appears in Lancet. 2020 Jan 30;:]. Lancet. 2020;395(10223):497‐506.
  3. Wang D, Hu B, Hu C, et al. Clinical Characteristics of 138 Hospitalized Patients With 2019 Novel Coronavirus-Infected Pneumonia in Wuhan, China [published online ahead of print, 2020 Feb 7]. JAMA. 2020;323(11):1061‐1069.
Real-time PCR solutions for SARS-CoV-2 research
GettyImages-1216476288-(1)
Our real-time PCR solutions combine sensitive and specific quantitation of nucleic acids with adaptable experimental design, flexible formats that enable affordable scalability and multi-target interrogation, easy-to-use workflows, and trusted support. Explore how are solutions for gene expression analysis, genotyping, and miRNA analysis can accelerate important discoveries across SARS-CoV-2 research areas.
Learn more ›
Translational research
The translational and pre-clinical research communities have responded quickly to SARS-CoV-2 outbreak by undertaking a variety of approaches to block viral infection and treat pathogenesis. Understanding disease mechanisms and the associated biomarkers through relevant experimental models, advanced cell engineering and protein modification techniques has expanded research capabilities. Thermo Fisher Scientific is committed to providing reliable and easy-to-use solutions, services and support to help meet the demanding needs of clinical and translational research, enabling researchers to transition rapidly from discovery research to translational settings and into the development of treatments.
Recombinant viruses provide excellent tools for the introduction of genetic material into a variety of host cells. We offer a wide range of tools including purification kits, plasmids, packaging systems and transduction kits and appropriate host cell lines for viral expression. In addition, we also offer tools for in vitro protein expression (also known as in vitro translation, cell-free protein expression, cell-free translation, or cell-free protein synthesis). Although in vitro expression is not practical for commercial large-scale recombinant protein production, it has a variety of features that make it considerably more useful and flexible for many research applications.
Experimental models: expression systems and cell-based assays
Invitrogen GeneArt Subcloning and Plasmid ServicesLearn more ›
Gibco Expi Protein Expression SystemsLearn more ›
Cell Free Protein Expression SystemsLearn more ›
Gibco LV-MAX Lentiviral Production systemLearn more ›
Expression systems
Catalog # Name Size Price (USD) Qty
A14635 Expi293™ Expression System Kit Each
2,690.65
Online Exclusive
3,165.00
Save 474.35 (15%)
A29133 ExpiCHO™ Expression System Kit Each
3,660.00
A38841 ExpiSf™ Expression System Starter Kit, 1 kit Each
2,360.00
A3711001 ExpiCHO™ Stable Production Medium Each
210.00
A35684 LV-MAX™ Lentiviral Production System Each
2,405.00
A35347 Viral Production Cells, 1 mL Each
1,306.00
A3583401 LV-MAX™ Production Medium, 1 L Each
305.83
Special offer
Online exclusive
Ends: 21-Jun-2025
340.00
Save 34.17 (10%)
A35348 LV-MAX™ Transfection Kit Each
1,630.98
Special offer
Online exclusive
Ends: 21-Jun-2025
1,786.00
Save 155.02 (9%)
31985088 Opti-MEM™ I Reduced Serum Medium, 10 x 500 mL Each
613.65
Online Exclusive
676.00
Save 62.35 (9%)
A43237 LV-MAX™ Lentiviral Packaging Mix Each
696.65
Online Exclusive
740.00
Save 43.35 (6%)
Quantigene
ProductCat No.
QuantiGene assaysLearn more ›
QuantiGene SingleplexRequest a quote ›
QuantiGene PlexRequest a quote ›
TaqMan Probe-based chemistry – For Research Use Only. Not for use in diagnostic procedures.
Catalog#Assay IDName
4331182Hs01085333_m1 / SARS-CoV-2ACE2
4331182Hs01122322_m1 / SARS-CoV-2TMPRSS2
4331182Hs00947439_m1 / SARS-CoV-2CTSB
4331182Hs00964650_m1 / SARS-CoV-2CTSL
4331182Hs00174265_m1 / HCoV-229EANPEP
4331182Hs00897386_m1 / MERS-CoVDPP4
4331182Hs00949382_m1 / InfluenzaST6GAL1
4331182Hs00920870_m1 / InfluenzaST3GAL4
Genome editing
To help you understand how the genome influences phenotype, our complete toolset offers trusted solutions for every step in the cell engineering workflow. Our collection of optimized, validated technology systems are designed to work together to eliminate the trial-and-error phase and help you get answers faster and with less effort.
Discover the genome editing tool ›
Easily create accurate and more successful knock-in experiments with TrueDesign Genome Editor. Our free design tool has an intuitive point-and-type interface that provides unrivaled flexibility and support for creating genome edits
CRISPR-mediated gene editing confirmation with Sanger sequencing
Because the repair process is not completely efficient or accurate in genome editing experiments, Sanger sequencing can be used to determine the efficiency of CRISPR-mediated genome editing in primary transformed cell cultures, and to determine successful editing events in secondary clones.
Applied Biosystems genetic analysis instruments and reagents provide:
  • Easy and unambiguous identification of single nucleotide polymorphisms (SNPs), sequence deletions, and insertions produced in a cellular population during gene editing
  • Minor Variant Finder Software and the cloud-based Sanger Variant Analysis module to determine the frequency of SNPs; compatible with Tracking of Indels by Decomposition (TIDE) software—a widely available tool for analyzing the efficiency of genome editing events
Learn about the genetic analysis tools available to confirm your gene editing experiments ›
GeneArt gene synthesis and services
Synthesis and protein engineeringGeneArt Gene Synthesis is a cost-effective method for obtaining customized DNA constructs that are 100% sequence accurate, allowing you to free up time and resources.
High-Q Strings DNA Fragmentsan economical and time-saving alternative to PCR that maintains the GeneArt Gene Synthesis benefits of flexibility, superior performance, and exceptional accuracy. GeneArt Strings and GeneArt High-Q Strings DNA fragments are compatible with any downstream cloning method.
GeneArt Gibson Assembly Cloning kitsGeneArt Gibson Assembly enables the construction of large and seamless constructs and in conjunction with GeneArt Gene and DNA fragment synthesis, allowing for fast and accurate construction of target Plasmid vectors.
DNA sequence amplification
Platinum SuperFi II DNA Polymerase—With >300x fidelity of Taq DNA polymerase and efficient amplification of DNA up to 40 kb, Platinum SuperFi II DNA Polymerase offers exceptional sequence accuracy and long-range amplification for cloning and mutagenesis of viral sequences.
Clone confirmation by Sanger sequencing
One of the most common applications of Sanger sequencing is the analysis of inserts subcloned into plasmids. Applied Biosystems BigDye chemistries are widely used for Sanger sequencing and are an integral part of plasmid sequencing workflows. The SeqStudio Genetic Analyzer has several features that offer benefits to researchers performing basic plasmid sequencing methods. The instrument is preloaded with sequencing modules optimized for short (<300 bp), medium (500 bp), and long (>600 bp) read lengths, and can also be customized on the instrument to meet specific needs. The cloud-based Sanger Quality Check application provides an intuitive set of tools to analyze sequencing traces.
Learn more about Sanger sequencing solutions for SARS-CoV-2 research ›
Transfection reagents and instruments
A broad range of Invitrogen transfection reagents and electroporation solutions optimized for efficiency, viability, and reproducibility and designed for effective delivery of nucleic acids. These include the Lipofectamine family of transfection reagents and the Neon electroporator.
Catalog # Name Size Price (USD) Qty
L3000008 Lipofectamine™ 3000 Transfection Reagent, 0.75 mL Each
563.65
Online Exclusive
598.00
Save 34.35 (6%)
GeneArt Gene-to-Protein Service
GeneArt Genes-to-Protein Service is a fast and efficient way to obtain correctly folded, native protein from transiently transfected mammalian cells. Starting with only the nucleotide sequence, we can provide purified protein within a few weeks. We clone your expression-optimized gene into one of our expression vectors, produce transfection-grade plasmid DNA, and then use one of our advanced expression systems to obtain high expression yields to support research needed for rapid protein production.
Protein purification
Easily extract, purify, clean up, and concentrate your proteins of interest with our kits, reagents, and devices. These products are optimized for a wide range of tissue and cell types and are compatible with a broad range of protein purification resins and formats, which offer more choices and better protein recovery. Desalt, buffer exchange, remove contaminants, and concentrate proteins with our secure and efficient devices and resins.
Crosslinking and labeling tools
We offer a wide range of products for the crosslinking, biotinylation, pegylation, and fluorescent labeling of proteins, antibodies, and peptides. Use our crosslinking or biotinylation reagent selection tool to find the optimal reagent for your application, or access our helpful technical handbook to help improve your bioconjugation results.
Antibody labeling
We offer a range of antibody labeling kits to choose from. Direct labeling of your primary antibody does provide two key advantages:
  • Multiple primary antibodies of the same isotype or derived from the same species can easily be used in the same experiment.
  • Primary antibodies directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding than labeled secondary antibodies.
Hight Yield Fast Performing MEGAscript in vitro transcription products
Catalog # Name Size Price (USD) Qty
AM1330 MEGAscript™ SP6 Transcription Kit, 40 Reactions Each
460.00
AM1334 MEGAscript™ T7 Transcription Kit, 40 Reactions Each
458.65
Online Exclusive
473.00
Save 14.35 (3%)
AM1333 MEGAscript™ T7 Transcription Kit, 25 Reactions Each
303.65
Online Exclusive
312.00
Save 8.35 (3%)
AMB13345 MEGAscript™ T7 Transcription Kit, 200 reactions Each
1,490.65
Online Exclusive
1,604.00
Save 113.35 (7%)
AM1338 MEGAscript™ T3 Transcription Kit Each
495.00
K0441 TranscriptAid T7 High Yield Transcription Kit Each
352.65
Online Exclusive
406.00
Save 53.35 (13%)
Capping in vitro transcription Products
Megaclear products
Catalog # Name Size Price (USD) Qty
AM1909 MEGAclear™-96 Transcription Clean-Up Kit Each
474.00
AM1908 MEGAclear™ Transcription Clean-Up Kit Each
191.00
Historically, researchers have relied upon 2D cell culture models, but growing evidence has highlighted the shortcoming of these model systems in predicting outcomes in more complex downstream animal models, settings required for therapeutic development. Increased understanding of cell to cell interactions and ex vivo growth conditions have allowed researchers to move into more complex ex vivo modeling cultures, such as 3D cell models, with the goal of providing a more natural cellular milieu. In addition, development of 3D cultures with primary cells can also be an avenue to assessing patient specific phenotypes and response to drug treatments.
3D models for studying SARS-CoV-2
Organoid and spheroid models are increasingly being used to more accurately mimic the in vivo cellular microenvironment, bringing more physiological relevance to experimental conditions. Generating organoid and spheroid models can be technically challenging, but once established can offer key insights for translational research projects, as they exhibit gene and protein expression signatures closer to those observed in vivo. 3D organoids are increasingly being used as infection models in lung/airways, gut, kidney, blood vessel, and other tissues to understand mechanisms of infection and immune response, identify key cellular targets, and test potential treatments (2). ​
References:
  1. Monteil V, Kwon H, Prado P, et al. Inhibition of SARS-CoV-2 Infections in Engineered Human Tissues Using Clinical-Grade Soluble Human ACE2. Cell. 2020;181(4):905‐913.e7.
  2. Ramani A, et al. SARS-CoV-2 targets cortical neurons of 3D human brain organoids and shows neurodegeneration-like effects. bioRxiv 2020.05.20.106575.
Primary cell culture system
Our systems allow you to closely mimic the in vivo state and generate more physiologically relevant data. Each lot of primary cells is performance tested for viability and growth potential.
Accurate cell counts are important for seeding cells for cell-based assays and prior to downstream analysis. The new Countess II FL Automated Cell Counter is a benchtop assay platform equipped with three-channels: brightfield and two optional fluorescence channels for cell counting, monitoring fluorescent protein expression, evaluating apoptosis, and measuring cell viability.
Find cell culture & transfection products for cell line development.
Catalog # Name Size Price (USD) Qty
C10228 Countess™ Cell Counting Chamber Slides, disposable, 50 Slides Each
143.65
Online Exclusive
153.00
Save 9.35 (6%)
ProductCat No.
Mammalian Cell Culture ReagentsLearn more ›
Genetic and phenotypic assessments are required to validate a cellular model in order to ensure its relevance and usability. Various cell types and sources that go through rounds of passaging, extensive engineering, or are exposed to experimental or environmental treatments and conditions need to be characterized. For example, in the case of adult primary stem cells, effects of passaging can influence the gene and protein expression patterns.
Authentication of human cell lines by Short Tandem Repeat analysis
Short tandem repeat (STR) genotyping using capillary electrophoresis is a simple, economical tool in verification of authenticity of human cell lines and primary stem cells and the assessment of the nature of known cell mixtures.  Misidentification of cell lines produces misleading results, confusion, and added costs to research. Journals and funding agencies now require researchers to prove that the cell lines they have used are authentic and have remained so over the course of a study. 
Analysis of highly variable STRs provides a highly specific genetic “fingerprint” of a cell line. Comparing a profile of alleles present at these highly variant loci to known, standardized samples of a cell line provides confidence that the cell line is authentic.  The Applied Biosystems CLA Identifiler Plus and Direct kits have been optimized to analyze 16 highly variant human STRs with amplification time of under three hours.  Applied Biosystems GeneMapper Software 6 and the cloud-based microsatellite analysis (MSA) software solutions facilitate analysis of STRs by making use of pre-established allelic ladders and sizing bin sets for the various STR alleles covered by the Identifiler kits.
Cell health tools for imaging and flow cytometry
Many cell health reagents have applications in both imaging and flow cytometry, such as those that measure toxicity, apoptosis, proliferation, viability, mitochondrial function, and membrane potential, etc.
Genotyping to verify genetic composition
For verification of genetic composition and sequence information of the cell models, PCR enzymes and master mixes serve as fast and efficient tools. Platinum Direct PCR Universal Master Mix offers direct amplification from cell samples without the need to purify DNA, shortening the workflow and reducing hands-on time. Platinum II Taq Hot-Start DNA Polymerase offers fast DNA synthesis and a simplified PCR protocol with a universal annealing temperature, for efficient amplification of target genomic sequences and their analysis by gel electrophoresis or Sanger sequencing.
Catalog # Name Size Price (USD) Qty
A42135 E-Gel™ Agarose Gels with SYBR™ Safe DNA Gel Stain, 2%, 10 Gels Each
162.00
10488091 E-Gel™ 1 Kb Plus Express DNA Ladder Each
238.65
Online Exclusive
248.00
Save 9.35 (4%)
G8300 E-Gel™ Power Snap Electrophoresis System Each
3,652.65
Online Exclusive
3,915.00
Save 262.35 (7%)
14966005 Platinum™ II Taq Hot-Start DNA Polymerase, 500 reactions Each
661.65
Online Exclusive
688.00
Save 26.35 (4%)
14000013 Platinum™ II Hot-Start PCR Master Mix (2X), 200 reactions Each
389.65
Online Exclusive
412.00
Save 22.35 (5%)
14001013 Platinum™ II Hot-Start Green PCR Master Mix (2X), 200 reactions Each
396.65
Online Exclusive
413.00
Save 16.35 (4%)
A44647100 Platinum™ Direct PCR Universal Master Mix, 100 reactions Each
143.65
Online Exclusive
149.00
Save 5.35 (4%)
Power Snap technology
Accelerate viral research with the Power Snap dry precast E-Gel agarose gel technology and run DNA samples in as little as 10 minutes. It is the only benchtop device that seamlessly integrates DNA sample separation and gel visualization into one workflow.
The majority of people diagnosed with SARS-CoV-2 infection recover from the infection. However, a small percentage develop complications. Use mass spectrometry-based proteomics and metabolomics workflows to understand the mechanism and help identify proteins and metabolites involved in the progression of SARS-CoV-2 infection.  Learn more ›
Virus detection by real-time PCR
Real-time PCR offers high sensitivity and specificity, fast time to results, and a wide dynamic range for precise detection and quantitation of nucleic acids. Known as the gold-standard for coronavirus detection and quantification, real-time PCR is a proven technology that can help accelerate life-saving discoveries across SARS-CoV-2 research areas.
Explore our real-time PCR solutions for SARS-CoV-2 research ›
Virus detection by mass spectrometry
The worldwide demand for testing of SARS-CoV-2 has created shortage in supplies for qRT-PCR, the current gold standard. Novel approaches that can overcome such shortages and increase testing capacity would be attractive. Mass spectrometry provides number of workflows that can circumvent such issues.
Learn more ›
Virus detection by capillary electrophoresis
Multiplexed qPCR solutions test for small numbers of pathogens, and its relatively small capacity can limit throughput when large numbers of targets or pathogens need to be detected. Fragment analysis by capillary electrophoresis (CE) can be used to test multiple pathogens associated with different syndromes in a single sample.  For example, a respiratory multiplex panel can detect viral and bacterial pathogens to help rule in/out common pathogens as the cause of infection.
Learn about a simple method for detecting SARS-CoV-2 viral sequences through fragment analysis-based target multiplexing solutions:
  • Application note: Multiplexed target fragment analysis for detection of viral pathogens including SARS-CoV-2
  •  Protocol: Detection of SARS-CoV-2 using fragment analysis
Reverse transcription loop-mediated isothermal amplification (RT-LAMP) protocol and application note for RNA-based amplification of viral pathogens, including  SARS-CoV-2
RT-LAMP is an easy technique for RNA-based amplification of viral pathogens. The RT-LAMP protocol includes reagents for fast and simple testing and surveillance of viral pathogens, including SARS-CoV-2, with a streamlined workflow that only needs a simple heat source to maintain a constant temperature. Several visualization methods are available to determine a positive RT-LAMP reaction, including endpoint and real-time (turbidity and dyes).
Catalog # Name Size Price (USD) Qty
EP0691 Bsm DNA Polymerase, large fragment (8 U/μL) Each
78.65
Online Exclusive
109.00
Save 30.35 (28%)
18090050 SuperScript™ IV Reverse Transcriptase, 10,000 Units Each
497.65
Online Exclusive
530.00
Save 32.35 (6%)
R0971 MgCl2 (magnesium chloride) (25 mM) Each
25.65
Online Exclusive
29.38
Save 3.73 (13%)
R0191 dNTP Mix (10 mM each), 0.2 mL Each
38.65
Online Exclusive
47.00
Save 8.35 (18%)
R0581 Water, nuclease-free, 4 x 1.25 mL Each
20.65
Online Exclusive
26.42
Save 5.77 (22%)
AM9890 DNAZap™ PCR DNA Degradation Solutions Each
134.65
Online Exclusive
141.00
Save 6.35 (5%)
AM9784 RNaseZap™ RNase Decontamination Solution, 4 L Each
919.65
Online Exclusive
962.00
Save 42.35 (4%)
AM9786 RNaseZap™ RNase Decontamination Wipes, 100 Sheets Each
122.65
Online Exclusive
128.00
Save 5.35 (4%)
10777019 RNaseOUT™ Recombinant Ribonuclease Inhibitor Each
255.00
S7567 SYBR™ Green I Nucleic Acid Gel Stain, 10,000X concentrate in DMSO, 1 mL Each
775.65
Online Exclusive
830.00
Save 54.35 (7%)
10488091 E-Gel™ 1 Kb Plus Express DNA Ladder Each
238.65
Online Exclusive
248.00
Save 9.35 (4%)
A42352 MagMAX™ Viral/Pathogen Nucleic Acid Isolation Kit, Up to 200 Preps Each
640.00
Therapeutic discovery & development
Therapeutic discovery and development is a complex process that can begin with screens, targets, or bioactive molecules that jumpstart the development workflow. From ready-to-use antiviral compounds to assays and complex libraries, we are here to support your research and development of potential therapeutics. In addition, our drug discovery and development team provides support, expertise, collaboration, and customization to meet your unique needs.
With over 40 antiviral compounds with proven activity, you can start your search for new antiviral compounds a few steps ahead of the curve, potentially speeding your time to results. A selection of antiviral compounds is shown below.
View our collection of chemical products ›
View real-time PCR solutions for SARS-CoV-2 research ›
Real-Time PCR can be used to quantify the virus before and after administration of antiviral compounds.
While the number of available full genomes produced by NGS can grow rapidly, datasets have historically faced challenges in the quality of the sequences. High-quality sequences provide coverage of entire genomes to help develop potential anti-viral therapies. In comparison to transposomic methods, the Invitrogen Collibri DNA Library Prep Kits for Illumina Systems consistently provide full coverage of RNA viruses, after reverse transcription, as well as host genomes. The Invitrogen Collibri Stranded RNA Kits for Illumina Systems with Human/Mouse/Rat Depletion attach helper adaptors directly to RNA, achieving robust coverage of the entire transcriptome.
Fragment analysis by capillary electrophoresis enables rapid screening for the presence or absence of SARS-CoV-2 RNA through a simple workflow.  Briefly, purified nucleic acid is mixed with a cocktail of fluorescent primers that are specific to the nucleoprotein (N protein), spike protein (S protein), and orf-1ab sequences in the SARS-CoV-2 genome. Primer pairs in the cocktail have been designed to generate well-separated, gene-specific amplicons. The sample–primer mixture is reverse transcribed and subjected to endpoint PCR in a single reaction. The resulting fragments are separated by size and detected using capillary electrophoresis.
The search for bioactive molecules in early-stage drug discovery using rapid compound screening and fragment-based design should not be like looking for a needle in a haystack. The Maybridge collection of screening compounds and fragment libraries offer a high degree of diversity in convenient and customizable formats with chemical properties designed to help accelerate your hit discovery.
Contact us ›
ProductCat No.
Antiviral library–over 8,000 compounds chosen to facilitate the discovery of novel chemical entities with profound viral activity and improved safety profilesLearn more ›
Maybridge HitDiscover–a pre-plated collection of over 53,000 screening compounds  that provide the diversity of the entire Maybridge collection.Learn more ›
Maybridge HitFinder–unique, pre-plated selections from the Maybridge collection chosen using  a clustering algorithm, all compounds fit Lipinski guidelines for "drug-likeness".Learn more ›
Maybridge HitCreator–14,000 compounds pre-plated in 96 or 384-well plates that represent the diversity of a 500,000 compound library distilled to 14,000 molecules. Learn more ›
The Maybridge Fragment Collection includes the overall fragment library as well as a fluor-,  19F-,  bromo-, and a pre-fragment library. Available as the entire library of 2,500 compounds or a core set of 1,000 and a supplemental set of 1,500, as well as customized sets  for your convenience.Learn more ›
Protein-protein interaction library–over 11,000 compounds selected through analysis of the PPI-relevant chemical spaceLearn more ›
GPCR library–over 10,000 compounds selected for screening against G-protein coupled receptorsLearn more ›
Kinase library–over 6,000 compounds chosen to facilitate the discovery of novel kinase inhibitors from our advanced collection of small molecules, fragments, and macrocyclesLearn more ›
With a variety of surfaces optimized to capture the biomolecule of your choice, a wide selection of microplates are available for your immunoassay development needs.
Discover how you can develop your own proprietary coronavirus assay leveraging our real-time PCR solutions for SARS-CoV-2 research.
A number of preclinical and clinical studies have been conducted investigating the cellular damage caused by SARS-CoV-2. For example, the immunomodulatory and reparative properties of mesenchymal stem cells could reduce mortality rate by reversing acute inflammation of lungs, inhibiting over-activation of immune cells, protecting lung alveoli epithelial cells, promoting tissue repair, preventing pulmonary fibrosis, or improving lung function. Thermo Fisher Scientific provides key resources to address challenges in the workflow for mesenchymal stem cell clinical research.
Learn more about cell therapy approaches and advancing your therapeutic mesenchymal stem cell therapy research with MSC media and reagents and custom media.
Human cells used in cell and gene therapy can be validated by Short tandem repeat (STR) genotyping using capillary electrophoresis. The Applied Biosystems CLA Identifiler Plus and Direct kits have been optimized to analyze 16 highly variant human STRs with amplification time of under three hours. Applied Biosystems GeneMapper Software 6 and the cloud-based microsatellite analysis (MSA) software solutions facilitate analysis of STRs by making use of pre-established allelic ladders and sizing bin sets for the various STR alleles covered by the Identifiler kits.
Learn more about how to use STR analysis to authenticate human cell lines ›
Catalog # Name Size Price (USD) Qty
A1067501 StemPro™ MSC SFM XenoFree Each
521.90
Special offer
Online exclusive
Ends: 21-Jun-2025
614.00
Save 92.10 (15%)
A1371301 CTS™ Synth-a-Freeze™ Medium Each
249.00
A1286001 CTS™ GlutaMAX™-I Supplement, 100 mL bottle Each
71.89
Special offer
Online exclusive
Ends: 21-Jun-2025
79.00
Save 7.11 (9%)
A4132602 CTS™ LV-MAX™ Transfection Kit, 1 Kit (For 10 L of LV Production) Each
19,920.00
CMAX00001 Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent, 0.1 mL Each
116.65
Online Exclusive
121.00
Save 4.35 (4%)
A36496 TrueCut™ Cas9 Protein v2, 10 μg Each
90.65
Online Exclusive
92.75
Save 2.10 (2%)
4460623 MycoSEQ™ Mycoplasma Detection Kit, 100 Reactions Each
4,195.00
A47695 CTS™ Rotea™ Counterflow Centrifugation System + 2-Year Service + IQOQ Each -
Catalog #Nameurl
4331182Hs01085333_m1 / SARS-CoV-2ACE2
4331182Hs01122322_m1 / SARS-CoV-2TMPRSS2
4331182Hs00947439_m1 / SARS-CoV-2CTSB
4331182Hs00964650_m1 / SARS-CoV-2CTSL
4331182Hs00174265_m1 / HCoV-229EANPEP
4331182Hs00897386_m1 / MERS-CoVDPP4
4331182Hs00949382_m1 / InfluenzaST6GAL1
4331182Hs00920870_m1 / InfluenzaST3GAL4
Bulk and specialty chemical services are available for custom synthesis, packaging, and testing, or for larger scale batches of candidate molecules. Learn more ›
For Research Use Only. Not for use in diagnostic procedures.